Assay Details
| Gene Symbol: IGSF23 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | NM_001205280.1 | NP_001192209.1 | 4 - 5 | 626 | 87 | |
| XM_017026323.1 | XP_016881812.1 | 4 - 5 | 800 | 87 | ||
| XM_017026325.1 | XP_016881814.1 | 3 - 4 | 646 | 87 | ||
| GenBank mRNA | BX112065.1 | - | 4 - 5 | 626 | 87 | |
More Information
Updated Mapping Information
This assay targets the same location as HTA transcript cluster(s) TC19000623.hg.1
Set Membership: |
|
MicroArray Probes:
Agilent
Affymetrix
ABI
Illumina
| Array Name | Probe IDs | Targets | Target Type |
|---|---|---|---|
| Agilent Human Whole Genome ChIP Tiling Array | A_17_P10972399 | - | - |
| Agilent Human Whole Genome ChIP Tiling Array | A_17_P10972374 | - | - |
| Agilent Human Whole Genome ChIP Tiling Array | A_17_P10972366 | - | - |
| Agilent Human Whole Genome ChIP Tiling Array | A_17_P10972404 | - | - |
| Agilent Human Whole Genome ChIP Tiling Array | A_17_P10972395 | - | - |
| Agilent Human Whole Genome ChIP Tiling Array | A_17_P31607261 | - | - |
| Agilent Human Whole Genome ChIP Tiling Array | A_17_P10972388 | - | - |
| Agilent Human Whole Genome ChIP Tiling Array | A_17_P10972384 | - | - |
| Agilent Human Whole Genome ChIP Tiling Array | A_17_P10972405 | - | - |
| Agilent Human Whole Genome ChIP Tiling Array | A_17_P10972390 | - | - |
| Agilent Human Whole Genome ChIP Tiling Array | A_17_P31607295 | - | - |
| Agilent Human Whole Genome ChIP Tiling Array | A_17_P10972398 | - | - |
| Agilent Human Whole Genome ChIP Tiling Array | A_17_P10972406 | - | - |
| Agilent Human Whole Genome ChIP Tiling Array | A_17_P10972369 | - | - |
| Agilent Human Whole Genome ChIP Tiling Array | A_17_P10972400 | - | - |
| Agilent Human Whole Genome ChIP Tiling Array | A_17_P10972407 | - | - |
| Agilent Human Whole Genome ChIP Tiling Array | A_17_P10972368 | - | - |
| Agilent Human Whole Genome ChIP Tiling Array | A_17_P17147062 | - | - |
| Agilent Human Whole Genome ChIP Tiling Array | A_17_P10972381 | - | - |
| Agilent Human Whole Genome ChIP Tiling Array | A_17_P10972380 | - | - |
| Agilent Human Whole Genome ChIP Tiling Array | A_17_P10972386 | - | - |
| Agilent Human Whole Genome ChIP Tiling Array | A_17_P10972375 | - | - |
| Agilent Human Whole Genome ChIP Tiling Array | A_17_P10972364 | - | - |
| Agilent Human Whole Genome ChIP Tiling Array | A_17_P10972370 | - | - |
| Agilent Human Whole Genome ChIP Tiling Array | A_17_P31607272 | - | - |
| Agilent Human Whole Genome ChIP Tiling Array | A_17_P10972401 | - | - |
| Agilent Human Whole Genome ChIP Tiling Array | A_17_P31607271 | - | - |
| Agilent Human Whole Genome ChIP Tiling Array | A_17_P31607297 | - | - |
| Agilent Human Whole Genome ChIP Tiling Array | A_17_P31607263 | - | - |
| Agilent Human Whole Genome ChIP Tiling Array | A_17_P31607290 | - | - |
| Agilent Human Whole Genome ChIP Tiling Array | A_17_P10972402 | - | - |
| Agilent Human Whole Genome ChIP Tiling Array | A_17_P10972376 | - | - |
| Agilent Human Whole Genome ChIP Tiling Array | A_17_P31607298 | - | - |
| Agilent Human Whole Genome ChIP Tiling Array | A_17_P10972385 | - | - |
| Agilent Human Whole Genome ChIP Tiling Array | A_17_P31607300 | - | - |
| Agilent Human Whole Genome ChIP Tiling Array | A_17_P10972379 | - | - |
| Agilent Human Whole Genome ChIP Tiling Array | A_17_P10972367 | - | - |
| Agilent Human Whole Genome ChIP Tiling Array | A_17_P31607280 | - | - |
| Agilent Human Whole Genome ChIP Tiling Array | A_17_P10972365 | - | - |
| Agilent Human Whole Genome ChIP Tiling Array | A_17_P10972377 | - | - |
| Agilent Human Whole Genome ChIP Tiling Array | A_17_P31607289 | - | - |
| Agilent Genotypic Designed Human Whole genome gene expression profiling microarray 4x44k | GT_44k_24_P750817 | - | - |
| Agilent Whole Human Genome Oligo Microarray 4x44K | 5318 | - | - |
| Agilent G4851A SurePrint G3 Hmn GE 8x60K Microarray | 51285 | - | - |
| Agilent Whole Human Genome Microarray 4x44K v2 | A_24_P750817 | - | - |
| Agilent Whole Human Genome Microarray 4x44K G4112F | A_24_P750817 | - | - |
| Agilent SurePrint G3 CGH array (4X180K) CCA_VUMC design AMADID#022522 | 6003 | - | - |
| Agilent Human CGH Whole Genome Microarray 1x1M (G4447A) | A_16_P21018214 | - | - |
| Agilent Human CGH Whole Genome Microarray 1x1M (G4447A) | A_16_P03454677 | - | - |
| Agilent Human CGH Whole Genome Microarray 1x1M (G4447A) | A_18_P21386813 | - | - |
| Agilent Human CGH Whole Genome Microarray 1x1M (G4447A) | A_16_P21018211 | - | - |
| Array Name | Probe IDs | Targets | Target Type |
|---|---|---|---|
| Affymetrix Human Transcriptome Array 2.0 | PSR19008645.hg.1 | ENST00000402988.1 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | PSR19008645.hg.1 | NM_001205280.1 | Refseq |
| Affymetrix Human Transcriptome Array 2.0 | PSR19008645.hg.1 | TC19000623.hg.1 | HTA TC |
| Affymetrix Human Transcriptome Array 2.0 | TC1900008271.hg.1 | ENST00000402988.5 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC1900008271.hg.1 | ENST00000402988.5 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC1900008271.hg.1 | ENST00000402988.5 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC1900008271.hg.1 | ENST00000402988.5 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC1900008271.hg.1 | ENST00000402988.5 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC1900008271.hg.1 | ENST00000402988.5 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC1900008271.hg.1 | ENST00000402988.5 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC1900008271.hg.1 | ENST00000402988.5 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC1900008271.hg.1 | ENST00000402988.5 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC1900008271.hg.1 | ENST00000428245.5 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC1900008271.hg.1 | ENST00000428245.5 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC1900008271.hg.1 | ENST00000428245.5 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC1900008271.hg.1 | ENST00000428245.5 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC1900008271.hg.1 | ENST00000428245.5 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC1900008271.hg.1 | ENST00000428245.5 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC1900008271.hg.1 | ENST00000428245.5 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC1900008271.hg.1 | ENST00000428245.5 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC1900008271.hg.1 | ENST00000428245.5 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC1900008271.hg.1 | ENST00000441389.1 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC1900008271.hg.1 | ENST00000441389.1 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC1900008271.hg.1 | ENST00000441389.1 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC1900008271.hg.1 | ENST00000441389.1 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC1900008271.hg.1 | ENST00000441389.1 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC1900008271.hg.1 | ENST00000441389.1 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC1900008271.hg.1 | ENST00000592507.1 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC1900008271.hg.1 | ENST00000592507.1 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC1900008271.hg.1 | ENST00000592507.1 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC1900008271.hg.1 | ENST00000592507.1 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC1900008271.hg.1 | NM_001205280.1 | Refseq |
| Affymetrix Human Transcriptome Array 2.0 | TC1900008271.hg.1 | NM_001205280.1 | Refseq |
| Affymetrix Human Transcriptome Array 2.0 | TC1900008271.hg.1 | NM_001205280.1 | Refseq |
| Affymetrix Human Transcriptome Array 2.0 | TC1900008271.hg.1 | NM_001205280.1 | Refseq |
| Affymetrix Human Transcriptome Array 2.0 | TC1900008271.hg.1 | NM_001205280.1 | Refseq |
| Affymetrix Human Transcriptome Array 2.0 | TC1900008271.hg.1 | NM_001205280.1 | Refseq |
| Affymetrix Human Transcriptome Array 2.0 | TC1900008271.hg.1 | NM_001205280.1 | Refseq |
| Affymetrix Human Transcriptome Array 2.0 | TC1900008271.hg.1 | NM_001205280.1 | Refseq |
| Affymetrix Human Transcriptome Array 2.0 | TC1900008271.hg.1 | NM_001205280.1 | Refseq |
| Affymetrix Human Transcriptome Array 2.0 | PSR19008644.hg.1 | ENST00000402988.1 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | PSR19008644.hg.1 | NM_001205280.1 | Refseq |
| Affymetrix Human Transcriptome Array 2.0 | PSR19008644.hg.1 | TC19000623.hg.1 | HTA TC |
| Affymetrix Human Clariom D Assay | TC1900008271.hg.1 | ENST00000402988.5 | Ensembl |
| Affymetrix Human Clariom D Assay | TC1900008271.hg.1 | ENST00000402988.5 | Ensembl |
| Affymetrix Human Clariom D Assay | TC1900008271.hg.1 | ENST00000402988.5 | Ensembl |
| Affymetrix Human Clariom D Assay | TC1900008271.hg.1 | ENST00000402988.5 | Ensembl |
| Affymetrix Human Clariom D Assay | TC1900008271.hg.1 | ENST00000402988.5 | Ensembl |
| Affymetrix Human Clariom D Assay | TC1900008271.hg.1 | ENST00000402988.5 | Ensembl |
| Affymetrix Human Clariom D Assay | TC1900008271.hg.1 | ENST00000402988.5 | Ensembl |
| Affymetrix Human Clariom D Assay | TC1900008271.hg.1 | ENST00000402988.5 | Ensembl |
| Affymetrix Human Clariom D Assay | TC1900008271.hg.1 | ENST00000402988.5 | Ensembl |
| Affymetrix Human Clariom D Assay | TC1900008271.hg.1 | ENST00000428245.5 | Ensembl |
| Affymetrix Human Clariom D Assay | TC1900008271.hg.1 | ENST00000428245.5 | Ensembl |
| Affymetrix Human Clariom D Assay | TC1900008271.hg.1 | ENST00000428245.5 | Ensembl |
| Affymetrix Human Clariom D Assay | TC1900008271.hg.1 | ENST00000428245.5 | Ensembl |
| Affymetrix Human Clariom D Assay | TC1900008271.hg.1 | ENST00000428245.5 | Ensembl |
| Affymetrix Human Clariom D Assay | TC1900008271.hg.1 | ENST00000428245.5 | Ensembl |
| Affymetrix Human Clariom D Assay | TC1900008271.hg.1 | ENST00000428245.5 | Ensembl |
| Affymetrix Human Clariom D Assay | TC1900008271.hg.1 | ENST00000428245.5 | Ensembl |
| Affymetrix Human Clariom D Assay | TC1900008271.hg.1 | ENST00000428245.5 | Ensembl |
| Affymetrix Human Clariom D Assay | TC1900008271.hg.1 | ENST00000441389.1 | Ensembl |
| Affymetrix Human Clariom D Assay | TC1900008271.hg.1 | ENST00000441389.1 | Ensembl |
| Affymetrix Human Clariom D Assay | TC1900008271.hg.1 | ENST00000441389.1 | Ensembl |
| Affymetrix Human Clariom D Assay | TC1900008271.hg.1 | ENST00000441389.1 | Ensembl |
| Affymetrix Human Clariom D Assay | TC1900008271.hg.1 | ENST00000441389.1 | Ensembl |
| Affymetrix Human Clariom D Assay | TC1900008271.hg.1 | ENST00000441389.1 | Ensembl |
| Affymetrix Human Clariom D Assay | TC1900008271.hg.1 | ENST00000592507.1 | Ensembl |
| Affymetrix Human Clariom D Assay | TC1900008271.hg.1 | ENST00000592507.1 | Ensembl |
| Affymetrix Human Clariom D Assay | TC1900008271.hg.1 | ENST00000592507.1 | Ensembl |
| Affymetrix Human Clariom D Assay | TC1900008271.hg.1 | ENST00000592507.1 | Ensembl |
| Affymetrix Human Clariom D Assay | TC1900008271.hg.1 | NM_001205280.1 | Refseq |
| Affymetrix Human Clariom D Assay | TC1900008271.hg.1 | NM_001205280.1 | Refseq |
| Affymetrix Human Clariom D Assay | TC1900008271.hg.1 | NM_001205280.1 | Refseq |
| Affymetrix Human Clariom D Assay | TC1900008271.hg.1 | NM_001205280.1 | Refseq |
| Affymetrix Human Clariom D Assay | TC1900008271.hg.1 | NM_001205280.1 | Refseq |
| Affymetrix Human Clariom D Assay | TC1900008271.hg.1 | NM_001205280.1 | Refseq |
| Affymetrix Human Clariom D Assay | TC1900008271.hg.1 | NM_001205280.1 | Refseq |
| Affymetrix Human Clariom D Assay | TC1900008271.hg.1 | NM_001205280.1 | Refseq |
| Affymetrix Human Clariom D Assay | TC1900008271.hg.1 | NM_001205280.1 | Refseq |
| Affymetrix Human Clariom D Assay | PSR1900163144.hg.1 | ENST00000402988.5 | Ensembl |
| Affymetrix Human Clariom D Assay | PSR1900163144.hg.1 | ENST00000428245.5 | Ensembl |
| Affymetrix Human Clariom D Assay | PSR1900163144.hg.1 | NM_001205280.1 | Refseq |
| Affymetrix Human Clariom D Assay | PSR1900163144.hg.1 | TC1900008271.hg.1 | Clariom D TC |
| Affymetrix Human Clariom D Assay | PSR1900163144.hg.1 | ENST00000402988.5 | Ensembl |
| Affymetrix Human Clariom D Assay | PSR1900163144.hg.1 | ENST00000428245.5 | Ensembl |
| Affymetrix Human Clariom D Assay | PSR1900163144.hg.1 | NM_001205280.1 | Refseq |
| Affymetrix Human Clariom D Assay | PSR1900163145.hg.1 | ENST00000402988.5 | Ensembl |
| Affymetrix Human Clariom D Assay | PSR1900163145.hg.1 | NM_001205280.1 | Refseq |
| Affymetrix Human Clariom D Assay | PSR1900163145.hg.1 | TC1900008271.hg.1 | Clariom D TC |
| Affymetrix Human Clariom D Assay | PSR1900163145.hg.1 | ENST00000402988.5 | Ensembl |
| Affymetrix Human Clariom D Assay | PSR1900163145.hg.1 | NM_001205280.1 | Refseq |
| Human Genome U95 Set | 60121_PM_at | - | - |
| Human Genome U95 Set | 60121_at | - | - |
| Array Name | Probe IDs | Targets | Target Type |
|---|---|---|---|
| ABI Human Genome Survey Microarray Version 2.0 | 156810 | - | - |
| Array Name | Probe IDs | Targets | Target Type |
|---|---|---|---|
| Illumina HumanWG-6 v2.0 expression beadchip | ILMN_1671865 | BX112065.1 | human_transcriptome |
| Illumina HumanWG-6 v2.0 expression beadchip | ILMN_1671865 | NM_001205280.1 | human_transcriptome |
| Illumina HumanWG-6 v2.0 expression beadchip | ILMN_1671865 | BC127928.1 | human_transcriptome |
| Illumina HumanWG-6 v2.0 expression beadchip | ILMN_1671865 | BC127929.1 | human_transcriptome |
| Illumina human-6 v2.0 expression beadchip | ILMN_1671865 | BX112065.1 | human_transcriptome |
| Illumina human-6 v2.0 expression beadchip | ILMN_1671865 | NM_001205280.1 | human_transcriptome |
| Illumina human-6 v2.0 expression beadchip | ILMN_1671865 | BC127928.1 | human_transcriptome |
| Illumina human-6 v2.0 expression beadchip | ILMN_1671865 | BC127929.1 | human_transcriptome |
| Illumina HumanWG-6 v3.0 expression beadchip | ILMN_1671865 | BX112065.1 | human_transcriptome |
| Illumina HumanWG-6 v3.0 expression beadchip | ILMN_1671865 | NM_001205280.1 | human_transcriptome |
| Illumina HumanWG-6 v3.0 expression beadchip | ILMN_1671865 | BC127928.1 | human_transcriptome |
| Illumina HumanWG-6 v3.0 expression beadchip | ILMN_1671865 | BC127929.1 | human_transcriptome |
| Illumina HumanHT-12 V3.0 expression beadchip | ILMN_1671865 | BX112065.1 | human_transcriptome |
| Illumina HumanHT-12 V3.0 expression beadchip | ILMN_1671865 | NM_001205280.1 | human_transcriptome |
| Illumina HumanHT-12 V3.0 expression beadchip | ILMN_1671865 | BC127928.1 | human_transcriptome |
| Illumina HumanHT-12 V3.0 expression beadchip | ILMN_1671865 | BC127929.1 | human_transcriptome |

IGSF23
Genomic Map