| Array Name |
Probe IDs |
Targets |
Target Type |
| Affymetrix Human Transcriptome Array 2.0 |
PSR01058105.hg.1 |
TC01003800.hg.1 |
HTA TC |
| Affymetrix Human Transcriptome Array 2.0 |
PSR01058105.hg.1 |
ENST00000366996.1 |
Ensembl |
| Affymetrix Human Transcriptome Array 2.0 |
PSR01058105.hg.1 |
ENST00000366997.4 |
Ensembl |
| Affymetrix Human Transcriptome Array 2.0 |
PSR01058105.hg.1 |
NM_014873.2 |
Refseq |
| Affymetrix Human Transcriptome Array 2.0 |
TC0100017223.hg.1 |
ENST00000366996.1 |
Ensembl |
| Affymetrix Human Transcriptome Array 2.0 |
TC0100017223.hg.1 |
ENST00000366996.1 |
Ensembl |
| Affymetrix Human Transcriptome Array 2.0 |
TC0100017223.hg.1 |
ENST00000366996.1 |
Ensembl |
| Affymetrix Human Transcriptome Array 2.0 |
TC0100017223.hg.1 |
ENST00000366996.1 |
Ensembl |
| Affymetrix Human Transcriptome Array 2.0 |
TC0100017223.hg.1 |
ENST00000366996.1 |
Ensembl |
| Affymetrix Human Transcriptome Array 2.0 |
TC0100017223.hg.1 |
ENST00000366996.1 |
Ensembl |
| Affymetrix Human Transcriptome Array 2.0 |
TC0100017223.hg.1 |
ENST00000366996.1 |
Ensembl |
| Affymetrix Human Transcriptome Array 2.0 |
TC0100017223.hg.1 |
ENST00000366996.1 |
Ensembl |
| Affymetrix Human Transcriptome Array 2.0 |
TC0100017223.hg.1 |
ENST00000366996.1 |
Ensembl |
| Affymetrix Human Transcriptome Array 2.0 |
TC0100017223.hg.1 |
ENST00000366996.1 |
Ensembl |
| Affymetrix Human Transcriptome Array 2.0 |
TC0100017223.hg.1 |
ENST00000366996.1 |
Ensembl |
| Affymetrix Human Transcriptome Array 2.0 |
TC0100017223.hg.1 |
ENST00000366996.1 |
Ensembl |
| Affymetrix Human Transcriptome Array 2.0 |
TC0100017223.hg.1 |
ENST00000366997.8 |
Ensembl |
| Affymetrix Human Transcriptome Array 2.0 |
TC0100017223.hg.1 |
ENST00000366997.8 |
Ensembl |
| Affymetrix Human Transcriptome Array 2.0 |
TC0100017223.hg.1 |
ENST00000366997.8 |
Ensembl |
| Affymetrix Human Transcriptome Array 2.0 |
TC0100017223.hg.1 |
ENST00000366997.8 |
Ensembl |
| Affymetrix Human Transcriptome Array 2.0 |
TC0100017223.hg.1 |
ENST00000366997.8 |
Ensembl |
| Affymetrix Human Transcriptome Array 2.0 |
TC0100017223.hg.1 |
ENST00000366997.8 |
Ensembl |
| Affymetrix Human Transcriptome Array 2.0 |
TC0100017223.hg.1 |
ENST00000366997.8 |
Ensembl |
| Affymetrix Human Transcriptome Array 2.0 |
TC0100017223.hg.1 |
ENST00000366997.8 |
Ensembl |
| Affymetrix Human Transcriptome Array 2.0 |
TC0100017223.hg.1 |
ENST00000366997.8 |
Ensembl |
| Affymetrix Human Transcriptome Array 2.0 |
TC0100017223.hg.1 |
ENST00000366997.8 |
Ensembl |
| Affymetrix Human Transcriptome Array 2.0 |
TC0100017223.hg.1 |
ENST00000366997.8 |
Ensembl |
| Affymetrix Human Transcriptome Array 2.0 |
TC0100017223.hg.1 |
ENST00000366997.8 |
Ensembl |
| Affymetrix Human Transcriptome Array 2.0 |
TC0100017223.hg.1 |
ENST00000366997.8 |
Ensembl |
| Affymetrix Human Transcriptome Array 2.0 |
TC0100017223.hg.1 |
ENST00000488600.1 |
Ensembl |
| Affymetrix Human Transcriptome Array 2.0 |
TC0100017223.hg.1 |
ENST00000488600.1 |
Ensembl |
| Affymetrix Human Transcriptome Array 2.0 |
TC0100017223.hg.1 |
ENST00000488600.1 |
Ensembl |
| Affymetrix Human Transcriptome Array 2.0 |
TC0100017223.hg.1 |
ENST00000488600.1 |
Ensembl |
| Affymetrix Human Transcriptome Array 2.0 |
TC0100017223.hg.1 |
ENST00000498690.1 |
Ensembl |
| Affymetrix Human Transcriptome Array 2.0 |
TC0100017223.hg.1 |
ENST00000498690.1 |
Ensembl |
| Affymetrix Human Transcriptome Array 2.0 |
TC0100017223.hg.1 |
ENST00000498690.1 |
Ensembl |
| Affymetrix Human Transcriptome Array 2.0 |
TC0100017223.hg.1 |
NM_014873.2 |
Refseq |
| Affymetrix Human Transcriptome Array 2.0 |
TC0100017223.hg.1 |
NM_014873.2 |
Refseq |
| Affymetrix Human Transcriptome Array 2.0 |
TC0100017223.hg.1 |
NM_014873.2 |
Refseq |
| Affymetrix Human Transcriptome Array 2.0 |
TC0100017223.hg.1 |
NM_014873.2 |
Refseq |
| Affymetrix Human Transcriptome Array 2.0 |
TC0100017223.hg.1 |
NM_014873.2 |
Refseq |
| Affymetrix Human Transcriptome Array 2.0 |
TC0100017223.hg.1 |
NM_014873.2 |
Refseq |
| Affymetrix Human Transcriptome Array 2.0 |
TC0100017223.hg.1 |
NM_014873.2 |
Refseq |
| Affymetrix Human Transcriptome Array 2.0 |
TC0100017223.hg.1 |
NM_014873.2 |
Refseq |
| Affymetrix Human Transcriptome Array 2.0 |
TC0100017223.hg.1 |
NM_014873.2 |
Refseq |
| Affymetrix Human Transcriptome Array 2.0 |
TC0100017223.hg.1 |
NM_014873.2 |
Refseq |
| Affymetrix Human Transcriptome Array 2.0 |
TC0100017223.hg.1 |
NM_014873.2 |
Refseq |
| Affymetrix Human Transcriptome Array 2.0 |
TC0100017223.hg.1 |
NM_014873.2 |
Refseq |
| Affymetrix Human Transcriptome Array 2.0 |
TC0100017223.hg.1 |
NM_014873.2 |
Refseq |
| Affymetrix Human Transcriptome Array 2.0 |
PSR01058106.hg.1 |
TC01003800.hg.1 |
HTA TC |
| Affymetrix Human Transcriptome Array 2.0 |
PSR01058106.hg.1 |
ENST00000366996.1 |
Ensembl |
| Affymetrix Human Transcriptome Array 2.0 |
PSR01058106.hg.1 |
ENST00000366997.4 |
Ensembl |
| Affymetrix Human Transcriptome Array 2.0 |
PSR01058106.hg.1 |
NM_014873.2 |
Refseq |
| Affymetrix Human Clariom D Assay |
PSR0100230413.hg.1 |
ENST00000366996.1 |
Ensembl |
| Affymetrix Human Clariom D Assay |
PSR0100230413.hg.1 |
ENST00000366997.8 |
Ensembl |
| Affymetrix Human Clariom D Assay |
PSR0100230413.hg.1 |
ENST00000498690.1 |
Ensembl |
| Affymetrix Human Clariom D Assay |
PSR0100230413.hg.1 |
NM_014873.2 |
Refseq |
| Affymetrix Human Clariom D Assay |
PSR0100230413.hg.1 |
ENST00000366996.1 |
Ensembl |
| Affymetrix Human Clariom D Assay |
PSR0100230413.hg.1 |
ENST00000366997.8 |
Ensembl |
| Affymetrix Human Clariom D Assay |
PSR0100230413.hg.1 |
ENST00000498690.1 |
Ensembl |
| Affymetrix Human Clariom D Assay |
PSR0100230413.hg.1 |
NM_014873.2 |
Refseq |
| Affymetrix Human Clariom D Assay |
PSR0100230413.hg.1 |
TC0100017223.hg.1 |
Clariom D TC |
| Affymetrix Human Clariom D Assay |
PSR0100230412.hg.1 |
ENST00000366996.1 |
Ensembl |
| Affymetrix Human Clariom D Assay |
PSR0100230412.hg.1 |
ENST00000366997.8 |
Ensembl |
| Affymetrix Human Clariom D Assay |
PSR0100230412.hg.1 |
NM_014873.2 |
Refseq |
| Affymetrix Human Clariom D Assay |
PSR0100230412.hg.1 |
ENST00000366996.1 |
Ensembl |
| Affymetrix Human Clariom D Assay |
PSR0100230412.hg.1 |
ENST00000366997.8 |
Ensembl |
| Affymetrix Human Clariom D Assay |
PSR0100230412.hg.1 |
NM_014873.2 |
Refseq |
| Affymetrix Human Clariom D Assay |
PSR0100230412.hg.1 |
TC0100017223.hg.1 |
Clariom D TC |
| Affymetrix Human Clariom D Assay |
TC0100017223.hg.1 |
ENST00000366996.1 |
Ensembl |
| Affymetrix Human Clariom D Assay |
TC0100017223.hg.1 |
ENST00000366996.1 |
Ensembl |
| Affymetrix Human Clariom D Assay |
TC0100017223.hg.1 |
ENST00000366996.1 |
Ensembl |
| Affymetrix Human Clariom D Assay |
TC0100017223.hg.1 |
ENST00000366996.1 |
Ensembl |
| Affymetrix Human Clariom D Assay |
TC0100017223.hg.1 |
ENST00000366996.1 |
Ensembl |
| Affymetrix Human Clariom D Assay |
TC0100017223.hg.1 |
ENST00000366996.1 |
Ensembl |
| Affymetrix Human Clariom D Assay |
TC0100017223.hg.1 |
ENST00000366996.1 |
Ensembl |
| Affymetrix Human Clariom D Assay |
TC0100017223.hg.1 |
ENST00000366996.1 |
Ensembl |
| Affymetrix Human Clariom D Assay |
TC0100017223.hg.1 |
ENST00000366996.1 |
Ensembl |
| Affymetrix Human Clariom D Assay |
TC0100017223.hg.1 |
ENST00000366996.1 |
Ensembl |
| Affymetrix Human Clariom D Assay |
TC0100017223.hg.1 |
ENST00000366996.1 |
Ensembl |
| Affymetrix Human Clariom D Assay |
TC0100017223.hg.1 |
ENST00000366996.1 |
Ensembl |
| Affymetrix Human Clariom D Assay |
TC0100017223.hg.1 |
ENST00000366997.8 |
Ensembl |
| Affymetrix Human Clariom D Assay |
TC0100017223.hg.1 |
ENST00000366997.8 |
Ensembl |
| Affymetrix Human Clariom D Assay |
TC0100017223.hg.1 |
ENST00000366997.8 |
Ensembl |
| Affymetrix Human Clariom D Assay |
TC0100017223.hg.1 |
ENST00000366997.8 |
Ensembl |
| Affymetrix Human Clariom D Assay |
TC0100017223.hg.1 |
ENST00000366997.8 |
Ensembl |
| Affymetrix Human Clariom D Assay |
TC0100017223.hg.1 |
ENST00000366997.8 |
Ensembl |
| Affymetrix Human Clariom D Assay |
TC0100017223.hg.1 |
ENST00000366997.8 |
Ensembl |
| Affymetrix Human Clariom D Assay |
TC0100017223.hg.1 |
ENST00000366997.8 |
Ensembl |
| Affymetrix Human Clariom D Assay |
TC0100017223.hg.1 |
ENST00000366997.8 |
Ensembl |
| Affymetrix Human Clariom D Assay |
TC0100017223.hg.1 |
ENST00000366997.8 |
Ensembl |
| Affymetrix Human Clariom D Assay |
TC0100017223.hg.1 |
ENST00000366997.8 |
Ensembl |
| Affymetrix Human Clariom D Assay |
TC0100017223.hg.1 |
ENST00000366997.8 |
Ensembl |
| Affymetrix Human Clariom D Assay |
TC0100017223.hg.1 |
ENST00000366997.8 |
Ensembl |
| Affymetrix Human Clariom D Assay |
TC0100017223.hg.1 |
ENST00000488600.1 |
Ensembl |
| Affymetrix Human Clariom D Assay |
TC0100017223.hg.1 |
ENST00000488600.1 |
Ensembl |
| Affymetrix Human Clariom D Assay |
TC0100017223.hg.1 |
ENST00000488600.1 |
Ensembl |
| Affymetrix Human Clariom D Assay |
TC0100017223.hg.1 |
ENST00000488600.1 |
Ensembl |
| Affymetrix Human Clariom D Assay |
TC0100017223.hg.1 |
ENST00000498690.1 |
Ensembl |
| Affymetrix Human Clariom D Assay |
TC0100017223.hg.1 |
ENST00000498690.1 |
Ensembl |
| Affymetrix Human Clariom D Assay |
TC0100017223.hg.1 |
ENST00000498690.1 |
Ensembl |
| Affymetrix Human Clariom D Assay |
TC0100017223.hg.1 |
NM_014873.2 |
Refseq |
| Affymetrix Human Clariom D Assay |
TC0100017223.hg.1 |
NM_014873.2 |
Refseq |
| Affymetrix Human Clariom D Assay |
TC0100017223.hg.1 |
NM_014873.2 |
Refseq |
| Affymetrix Human Clariom D Assay |
TC0100017223.hg.1 |
NM_014873.2 |
Refseq |
| Affymetrix Human Clariom D Assay |
TC0100017223.hg.1 |
NM_014873.2 |
Refseq |
| Affymetrix Human Clariom D Assay |
TC0100017223.hg.1 |
NM_014873.2 |
Refseq |
| Affymetrix Human Clariom D Assay |
TC0100017223.hg.1 |
NM_014873.2 |
Refseq |
| Affymetrix Human Clariom D Assay |
TC0100017223.hg.1 |
NM_014873.2 |
Refseq |
| Affymetrix Human Clariom D Assay |
TC0100017223.hg.1 |
NM_014873.2 |
Refseq |
| Affymetrix Human Clariom D Assay |
TC0100017223.hg.1 |
NM_014873.2 |
Refseq |
| Affymetrix Human Clariom D Assay |
TC0100017223.hg.1 |
NM_014873.2 |
Refseq |
| Affymetrix Human Clariom D Assay |
TC0100017223.hg.1 |
NM_014873.2 |
Refseq |
| Affymetrix Human Clariom D Assay |
TC0100017223.hg.1 |
NM_014873.2 |
Refseq |