Assay Details
| Gene Symbol: PCTP | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | NM_001102402.2 | NP_001095872.1 | 1 - 2 | 370 | 76 | |
| XM_017024913.1 | XP_016880402.1 | 1 - 2 | 345 | 76 | ||
| GenBank mRNA | AK058120.1 | - | 1 - 2 | 196 | 76 | |
| BC005112.1 | - | 1 - 2 | 75 | 76 | ||
| BM549250.1 | - | 1 - 2 | 370 | 76 | ||
More Information
Updated Mapping Information
This assay targets the same location as HTA transcript cluster(s) TC17000695.hg.1
Additional Information:
This assay is in the 5'UTR region of the NM_001102402.2,XM_017024913.1 transcripts.
Set Membership: |
|
Gene Ontology Categories:
MicroArray Probes:
Affymetrix
ABI
| Array Name | Probe IDs | Targets | Target Type |
|---|---|---|---|
| Affymetrix Human Transcriptome Array 2.0 | TC1700008342.hg.1 | ENST00000268896.9 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC1700008342.hg.1 | ENST00000268896.9 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC1700008342.hg.1 | ENST00000268896.9 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC1700008342.hg.1 | ENST00000268896.9 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC1700008342.hg.1 | ENST00000268896.9 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC1700008342.hg.1 | ENST00000268896.9 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC1700008342.hg.1 | ENST00000268896.9 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC1700008342.hg.1 | ENST00000268896.9 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC1700008342.hg.1 | ENST00000268896.9 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC1700008342.hg.1 | ENST00000268896.9 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC1700008342.hg.1 | ENST00000325214.10 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC1700008342.hg.1 | ENST00000325214.10 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC1700008342.hg.1 | ENST00000325214.10 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC1700008342.hg.1 | ENST00000325214.10 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC1700008342.hg.1 | ENST00000325214.10 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC1700008342.hg.1 | ENST00000325214.10 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC1700008342.hg.1 | ENST00000325214.10 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC1700008342.hg.1 | ENST00000325214.10 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC1700008342.hg.1 | ENST00000417982.6 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC1700008342.hg.1 | ENST00000417982.6 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC1700008342.hg.1 | ENST00000417982.6 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC1700008342.hg.1 | ENST00000417982.6 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC1700008342.hg.1 | ENST00000417982.6 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC1700008342.hg.1 | ENST00000417982.6 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC1700008342.hg.1 | ENST00000417982.6 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC1700008342.hg.1 | ENST00000417982.6 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC1700008342.hg.1 | ENST00000417982.6 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC1700008342.hg.1 | ENST00000417982.6 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC1700008342.hg.1 | ENST00000417982.6 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC1700008342.hg.1 | ENST00000571489.5 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC1700008342.hg.1 | ENST00000571489.5 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC1700008342.hg.1 | ENST00000571489.5 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC1700008342.hg.1 | ENST00000571489.5 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC1700008342.hg.1 | ENST00000571489.5 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC1700008342.hg.1 | ENST00000571489.5 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC1700008342.hg.1 | ENST00000572536.1 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC1700008342.hg.1 | ENST00000572536.1 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC1700008342.hg.1 | ENST00000572536.1 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC1700008342.hg.1 | ENST00000572536.1 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC1700008342.hg.1 | ENST00000572536.1 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC1700008342.hg.1 | ENST00000573500.5 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC1700008342.hg.1 | ENST00000573500.5 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC1700008342.hg.1 | ENST00000573500.5 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC1700008342.hg.1 | ENST00000573500.5 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC1700008342.hg.1 | ENST00000573500.5 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC1700008342.hg.1 | ENST00000573500.5 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC1700008342.hg.1 | ENST00000573500.5 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC1700008342.hg.1 | ENST00000573500.5 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC1700008342.hg.1 | ENST00000576183.5 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC1700008342.hg.1 | ENST00000576183.5 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC1700008342.hg.1 | ENST00000576183.5 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC1700008342.hg.1 | ENST00000576183.5 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC1700008342.hg.1 | ENST00000576183.5 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC1700008342.hg.1 | ENST00000576183.5 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC1700008342.hg.1 | ENST00000576183.5 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC1700008342.hg.1 | ENST00000576183.5 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC1700008342.hg.1 | ENST00000576183.5 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC1700008342.hg.1 | ENST00000576183.5 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC1700008342.hg.1 | ENST00000576221.1 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC1700008342.hg.1 | ENST00000576221.1 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC1700008342.hg.1 | ENST00000576221.1 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC1700008342.hg.1 | ENST00000576221.1 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC1700008342.hg.1 | ENST00000576221.1 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC1700008342.hg.1 | NM_001102402.2 | Refseq |
| Affymetrix Human Transcriptome Array 2.0 | TC1700008342.hg.1 | NM_001102402.2 | Refseq |
| Affymetrix Human Transcriptome Array 2.0 | TC1700008342.hg.1 | NM_001102402.2 | Refseq |
| Affymetrix Human Transcriptome Array 2.0 | TC1700008342.hg.1 | NM_001102402.2 | Refseq |
| Affymetrix Human Transcriptome Array 2.0 | TC1700008342.hg.1 | NM_001102402.2 | Refseq |
| Affymetrix Human Transcriptome Array 2.0 | TC1700008342.hg.1 | NM_001102402.2 | Refseq |
| Affymetrix Human Transcriptome Array 2.0 | TC1700008342.hg.1 | NM_001102402.2 | Refseq |
| Affymetrix Human Transcriptome Array 2.0 | TC1700008342.hg.1 | NM_001102402.2 | Refseq |
| Affymetrix Human Transcriptome Array 2.0 | TC1700008342.hg.1 | NM_001102402.2 | Refseq |
| Affymetrix Human Transcriptome Array 2.0 | TC1700008342.hg.1 | NM_001102402.2 | Refseq |
| Affymetrix Human Transcriptome Array 2.0 | TC1700008342.hg.1 | NM_021213.3 | Refseq |
| Affymetrix Human Transcriptome Array 2.0 | TC1700008342.hg.1 | NM_021213.3 | Refseq |
| Affymetrix Human Transcriptome Array 2.0 | TC1700008342.hg.1 | NM_021213.3 | Refseq |
| Affymetrix Human Transcriptome Array 2.0 | TC1700008342.hg.1 | NM_021213.3 | Refseq |
| Affymetrix Human Transcriptome Array 2.0 | TC1700008342.hg.1 | NM_021213.3 | Refseq |
| Affymetrix Human Transcriptome Array 2.0 | TC1700008342.hg.1 | NM_021213.3 | Refseq |
| Affymetrix Human Transcriptome Array 2.0 | TC1700008342.hg.1 | NM_021213.3 | Refseq |
| Affymetrix Human Transcriptome Array 2.0 | TC1700008342.hg.1 | NM_021213.3 | Refseq |
| Affymetrix Human Transcriptome Array 2.0 | TC1700008342.hg.1 | NM_021213.3 | Refseq |
| Affymetrix Human Transcriptome Array 2.0 | TC1700008342.hg.1 | NM_021213.3 | Refseq |
| Affymetrix Human Transcriptome Array 2.0 | PSR17009114.hg.1 | ENST00000325214.5 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | PSR17009114.hg.1 | ENST00000417982.1 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | PSR17009114.hg.1 | NM_001102402.2 | Refseq |
| Affymetrix Human Transcriptome Array 2.0 | PSR17009114.hg.1 | TC17000695.hg.1 | HTA TC |
| Affymetrix Human Transcriptome Array 2.0 | PSR17009113.hg.1 | ENST00000417982.1 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | PSR17009113.hg.1 | NM_001102402.2 | Refseq |
| Affymetrix Human Transcriptome Array 2.0 | PSR17009113.hg.1 | TC17000695.hg.1 | HTA TC |
| Affymetrix Human Transcriptome Array 2.0 | PSR17009115.hg.1 | ENST00000268896.4 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | PSR17009115.hg.1 | ENST00000325214.5 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | PSR17009115.hg.1 | ENST00000417982.1 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | PSR17009115.hg.1 | NM_001102402.2 | Refseq |
| Affymetrix Human Transcriptome Array 2.0 | PSR17009115.hg.1 | NM_021213.3 | Refseq |
| Affymetrix Human Transcriptome Array 2.0 | PSR17009115.hg.1 | TC17000695.hg.1 | HTA TC |
| Affymetrix Human Clariom D Assay | PSR1700162174.hg.1 | ENST00000325214.10 | Ensembl |
| Affymetrix Human Clariom D Assay | PSR1700162174.hg.1 | ENST00000417982.6 | Ensembl |
| Affymetrix Human Clariom D Assay | PSR1700162174.hg.1 | NM_001102402.2 | Refseq |
| Affymetrix Human Clariom D Assay | PSR1700162174.hg.1 | ENST00000325214.10 | Ensembl |
| Affymetrix Human Clariom D Assay | PSR1700162174.hg.1 | ENST00000417982.6 | Ensembl |
| Affymetrix Human Clariom D Assay | PSR1700162174.hg.1 | NM_001102402.2 | Refseq |
| Affymetrix Human Clariom D Assay | PSR1700162174.hg.1 | TC1700008342.hg.1 | Clariom D TC |
| Affymetrix Human Clariom D Assay | PSR1700162175.hg.1 | ENST00000268896.9 | Ensembl |
| Affymetrix Human Clariom D Assay | PSR1700162175.hg.1 | ENST00000325214.10 | Ensembl |
| Affymetrix Human Clariom D Assay | PSR1700162175.hg.1 | ENST00000417982.6 | Ensembl |
| Affymetrix Human Clariom D Assay | PSR1700162175.hg.1 | ENST00000573500.5 | Ensembl |
| Affymetrix Human Clariom D Assay | PSR1700162175.hg.1 | ENST00000576183.5 | Ensembl |
| Affymetrix Human Clariom D Assay | PSR1700162175.hg.1 | NM_001102402.2 | Refseq |
| Affymetrix Human Clariom D Assay | PSR1700162175.hg.1 | NM_021213.3 | Refseq |
| Affymetrix Human Clariom D Assay | PSR1700162175.hg.1 | ENST00000268896.9 | Ensembl |
| Affymetrix Human Clariom D Assay | PSR1700162175.hg.1 | ENST00000325214.10 | Ensembl |
| Affymetrix Human Clariom D Assay | PSR1700162175.hg.1 | ENST00000417982.6 | Ensembl |
| Affymetrix Human Clariom D Assay | PSR1700162175.hg.1 | ENST00000573500.5 | Ensembl |
| Affymetrix Human Clariom D Assay | PSR1700162175.hg.1 | ENST00000576183.5 | Ensembl |
| Affymetrix Human Clariom D Assay | PSR1700162175.hg.1 | NM_001102402.2 | Refseq |
| Affymetrix Human Clariom D Assay | PSR1700162175.hg.1 | NM_021213.3 | Refseq |
| Affymetrix Human Clariom D Assay | PSR1700162175.hg.1 | TC1700008342.hg.1 | Clariom D TC |
| Affymetrix Human Clariom D Assay | TC1700008342.hg.1 | ENST00000268896.9 | Ensembl |
| Affymetrix Human Clariom D Assay | TC1700008342.hg.1 | ENST00000268896.9 | Ensembl |
| Affymetrix Human Clariom D Assay | TC1700008342.hg.1 | ENST00000268896.9 | Ensembl |
| Affymetrix Human Clariom D Assay | TC1700008342.hg.1 | ENST00000268896.9 | Ensembl |
| Affymetrix Human Clariom D Assay | TC1700008342.hg.1 | ENST00000268896.9 | Ensembl |
| Affymetrix Human Clariom D Assay | TC1700008342.hg.1 | ENST00000268896.9 | Ensembl |
| Affymetrix Human Clariom D Assay | TC1700008342.hg.1 | ENST00000268896.9 | Ensembl |
| Affymetrix Human Clariom D Assay | TC1700008342.hg.1 | ENST00000268896.9 | Ensembl |
| Affymetrix Human Clariom D Assay | TC1700008342.hg.1 | ENST00000268896.9 | Ensembl |
| Affymetrix Human Clariom D Assay | TC1700008342.hg.1 | ENST00000268896.9 | Ensembl |
| Affymetrix Human Clariom D Assay | TC1700008342.hg.1 | ENST00000325214.10 | Ensembl |
| Affymetrix Human Clariom D Assay | TC1700008342.hg.1 | ENST00000325214.10 | Ensembl |
| Affymetrix Human Clariom D Assay | TC1700008342.hg.1 | ENST00000325214.10 | Ensembl |
| Affymetrix Human Clariom D Assay | TC1700008342.hg.1 | ENST00000325214.10 | Ensembl |
| Affymetrix Human Clariom D Assay | TC1700008342.hg.1 | ENST00000325214.10 | Ensembl |
| Affymetrix Human Clariom D Assay | TC1700008342.hg.1 | ENST00000325214.10 | Ensembl |
| Affymetrix Human Clariom D Assay | TC1700008342.hg.1 | ENST00000325214.10 | Ensembl |
| Affymetrix Human Clariom D Assay | TC1700008342.hg.1 | ENST00000325214.10 | Ensembl |
| Affymetrix Human Clariom D Assay | TC1700008342.hg.1 | ENST00000417982.6 | Ensembl |
| Affymetrix Human Clariom D Assay | TC1700008342.hg.1 | ENST00000417982.6 | Ensembl |
| Affymetrix Human Clariom D Assay | TC1700008342.hg.1 | ENST00000417982.6 | Ensembl |
| Affymetrix Human Clariom D Assay | TC1700008342.hg.1 | ENST00000417982.6 | Ensembl |
| Affymetrix Human Clariom D Assay | TC1700008342.hg.1 | ENST00000417982.6 | Ensembl |
| Affymetrix Human Clariom D Assay | TC1700008342.hg.1 | ENST00000417982.6 | Ensembl |
| Affymetrix Human Clariom D Assay | TC1700008342.hg.1 | ENST00000417982.6 | Ensembl |
| Affymetrix Human Clariom D Assay | TC1700008342.hg.1 | ENST00000417982.6 | Ensembl |
| Affymetrix Human Clariom D Assay | TC1700008342.hg.1 | ENST00000417982.6 | Ensembl |
| Affymetrix Human Clariom D Assay | TC1700008342.hg.1 | ENST00000417982.6 | Ensembl |
| Affymetrix Human Clariom D Assay | TC1700008342.hg.1 | ENST00000417982.6 | Ensembl |
| Affymetrix Human Clariom D Assay | TC1700008342.hg.1 | ENST00000571489.5 | Ensembl |
| Affymetrix Human Clariom D Assay | TC1700008342.hg.1 | ENST00000571489.5 | Ensembl |
| Affymetrix Human Clariom D Assay | TC1700008342.hg.1 | ENST00000571489.5 | Ensembl |
| Affymetrix Human Clariom D Assay | TC1700008342.hg.1 | ENST00000571489.5 | Ensembl |
| Affymetrix Human Clariom D Assay | TC1700008342.hg.1 | ENST00000571489.5 | Ensembl |
| Affymetrix Human Clariom D Assay | TC1700008342.hg.1 | ENST00000571489.5 | Ensembl |
| Affymetrix Human Clariom D Assay | TC1700008342.hg.1 | ENST00000572536.1 | Ensembl |
| Affymetrix Human Clariom D Assay | TC1700008342.hg.1 | ENST00000572536.1 | Ensembl |
| Affymetrix Human Clariom D Assay | TC1700008342.hg.1 | ENST00000572536.1 | Ensembl |
| Affymetrix Human Clariom D Assay | TC1700008342.hg.1 | ENST00000572536.1 | Ensembl |
| Affymetrix Human Clariom D Assay | TC1700008342.hg.1 | ENST00000572536.1 | Ensembl |
| Affymetrix Human Clariom D Assay | TC1700008342.hg.1 | ENST00000573500.5 | Ensembl |
| Affymetrix Human Clariom D Assay | TC1700008342.hg.1 | ENST00000573500.5 | Ensembl |
| Affymetrix Human Clariom D Assay | TC1700008342.hg.1 | ENST00000573500.5 | Ensembl |
| Affymetrix Human Clariom D Assay | TC1700008342.hg.1 | ENST00000573500.5 | Ensembl |
| Affymetrix Human Clariom D Assay | TC1700008342.hg.1 | ENST00000573500.5 | Ensembl |
| Affymetrix Human Clariom D Assay | TC1700008342.hg.1 | ENST00000573500.5 | Ensembl |
| Affymetrix Human Clariom D Assay | TC1700008342.hg.1 | ENST00000573500.5 | Ensembl |
| Affymetrix Human Clariom D Assay | TC1700008342.hg.1 | ENST00000573500.5 | Ensembl |
| Affymetrix Human Clariom D Assay | TC1700008342.hg.1 | ENST00000576183.5 | Ensembl |
| Affymetrix Human Clariom D Assay | TC1700008342.hg.1 | ENST00000576183.5 | Ensembl |
| Affymetrix Human Clariom D Assay | TC1700008342.hg.1 | ENST00000576183.5 | Ensembl |
| Affymetrix Human Clariom D Assay | TC1700008342.hg.1 | ENST00000576183.5 | Ensembl |
| Affymetrix Human Clariom D Assay | TC1700008342.hg.1 | ENST00000576183.5 | Ensembl |
| Affymetrix Human Clariom D Assay | TC1700008342.hg.1 | ENST00000576183.5 | Ensembl |
| Affymetrix Human Clariom D Assay | TC1700008342.hg.1 | ENST00000576183.5 | Ensembl |
| Affymetrix Human Clariom D Assay | TC1700008342.hg.1 | ENST00000576183.5 | Ensembl |
| Affymetrix Human Clariom D Assay | TC1700008342.hg.1 | ENST00000576183.5 | Ensembl |
| Affymetrix Human Clariom D Assay | TC1700008342.hg.1 | ENST00000576183.5 | Ensembl |
| Affymetrix Human Clariom D Assay | TC1700008342.hg.1 | ENST00000576221.1 | Ensembl |
| Affymetrix Human Clariom D Assay | TC1700008342.hg.1 | ENST00000576221.1 | Ensembl |
| Affymetrix Human Clariom D Assay | TC1700008342.hg.1 | ENST00000576221.1 | Ensembl |
| Affymetrix Human Clariom D Assay | TC1700008342.hg.1 | ENST00000576221.1 | Ensembl |
| Affymetrix Human Clariom D Assay | TC1700008342.hg.1 | ENST00000576221.1 | Ensembl |
| Affymetrix Human Clariom D Assay | TC1700008342.hg.1 | NM_001102402.2 | Refseq |
| Affymetrix Human Clariom D Assay | TC1700008342.hg.1 | NM_001102402.2 | Refseq |
| Affymetrix Human Clariom D Assay | TC1700008342.hg.1 | NM_001102402.2 | Refseq |
| Affymetrix Human Clariom D Assay | TC1700008342.hg.1 | NM_001102402.2 | Refseq |
| Affymetrix Human Clariom D Assay | TC1700008342.hg.1 | NM_001102402.2 | Refseq |
| Affymetrix Human Clariom D Assay | TC1700008342.hg.1 | NM_001102402.2 | Refseq |
| Affymetrix Human Clariom D Assay | TC1700008342.hg.1 | NM_001102402.2 | Refseq |
| Affymetrix Human Clariom D Assay | TC1700008342.hg.1 | NM_001102402.2 | Refseq |
| Affymetrix Human Clariom D Assay | TC1700008342.hg.1 | NM_001102402.2 | Refseq |
| Affymetrix Human Clariom D Assay | TC1700008342.hg.1 | NM_001102402.2 | Refseq |
| Affymetrix Human Clariom D Assay | TC1700008342.hg.1 | NM_021213.3 | Refseq |
| Affymetrix Human Clariom D Assay | TC1700008342.hg.1 | NM_021213.3 | Refseq |
| Affymetrix Human Clariom D Assay | TC1700008342.hg.1 | NM_021213.3 | Refseq |
| Affymetrix Human Clariom D Assay | TC1700008342.hg.1 | NM_021213.3 | Refseq |
| Affymetrix Human Clariom D Assay | TC1700008342.hg.1 | NM_021213.3 | Refseq |
| Affymetrix Human Clariom D Assay | TC1700008342.hg.1 | NM_021213.3 | Refseq |
| Affymetrix Human Clariom D Assay | TC1700008342.hg.1 | NM_021213.3 | Refseq |
| Affymetrix Human Clariom D Assay | TC1700008342.hg.1 | NM_021213.3 | Refseq |
| Affymetrix Human Clariom D Assay | TC1700008342.hg.1 | NM_021213.3 | Refseq |
| Affymetrix Human Clariom D Assay | TC1700008342.hg.1 | NM_021213.3 | Refseq |
| Affymetrix Human Clariom D Assay | PSR1700162173.hg.1 | ENST00000417982.6 | Ensembl |
| Affymetrix Human Clariom D Assay | PSR1700162173.hg.1 | NM_001102402.2 | Refseq |
| Affymetrix Human Clariom D Assay | PSR1700162173.hg.1 | ENST00000417982.6 | Ensembl |
| Affymetrix Human Clariom D Assay | PSR1700162173.hg.1 | NM_001102402.2 | Refseq |
| Affymetrix Human Clariom D Assay | PSR1700162173.hg.1 | TC1700008342.hg.1 | Clariom D TC |
| Array Name | Probe IDs | Targets | Target Type |
|---|---|---|---|
| ABI Human Genome Survey Microarray Version 2.0 | 202139 | - | - |

PCTP
Genomic Map