Assay Details
| Gene Symbol: TSPAN18 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | XM_005253217.3 | XP_005253274.1 | 4 - 5 | 413 | 64 | |
| XM_006718372.3 | XP_006718435.1 | 4 - 5 | 500 | 64 | ||
| XM_017018530.1 | XP_016874019.1 | 4 - 5 | 446 | 64 | ||
More Information
Additional Information:
This assay is in the 5'UTR region of the XM_005253217.3, XM_006718372.3, XM_017018530.1 transcripts.
Set Membership: |
|

TSPAN18
Genomic Map