Assay Details
| Gene Symbol: CRY2 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | NM_001127457.2 | NP_001120929.1 | 8 - 9 | 1294 | 137 | |
| NM_021117.3 | NP_066940.2 | 8 - 9 | 1430 | 137 | ||
| GenBank mRNA | AB014558.1 | - | 8 - 9 | 1334 | 137 | |
| AK291260.1 | - | 8 - 9 | 1361 | 137 | ||
| AK294904.1 | - | 8 - 9 | 1288 | 137 | ||
| AK295627.1 | - | 4 - 5 | 830 | 137 | ||
| AK302865.1 | - | 8 - 9 | 1430 | 137 | ||
| BC035161.1 | - | 10 - 11 | 1693 | 5265444 | 137 | |
| BC041814.1 | - | 8 - 9 | 1358 | 5266781 | 137 | |
| EU219622.1 | - | 8 - 9 | 1353 | 137 | ||
More Information
Updated Mapping Information
This assay targets the same location as HTA transcript cluster(s) TC11000388.hg.1
Set Membership: |
|
Matching Ambion siRNAs:
Panther Classification:
Gene Ontology Categories:
Function(s)
Process(es)
negative regulation of transcription from RNA polymerase II promoter
photoreactive repair
transcription, DNA-templated
circadian rhythm
blue light signaling pathway
protein-chromophore linkage
negative regulation of phosphoprotein phosphatase activity
circadian regulation of gene expression
glucose homeostasis
regulation of circadian rhythm
negative regulation of circadian rhythm
entrainment of circadian clock by photoperiod
negative regulation of transcription, DNA-templated
regulation of sodium-dependent phosphate transport
negative regulation of glucocorticoid receptor signaling pathway
transcription regulatory region sequence-specific DNA binding
transcription factor activity, transcription factor binding
DNA binding
damaged DNA binding
single-stranded DNA binding
deoxyribodipyrimidine photo-lyase activity
DNA (6-4) photolyase activity
protein binding
blue light photoreceptor activity
phosphatase binding
ubiquitin binding
FAD binding
negative regulation of transcription from RNA polymerase II promoter
photoreactive repair
transcription, DNA-templated
circadian rhythm
blue light signaling pathway
protein-chromophore linkage
negative regulation of phosphoprotein phosphatase activity
circadian regulation of gene expression
glucose homeostasis
regulation of circadian rhythm
negative regulation of circadian rhythm
entrainment of circadian clock by photoperiod
negative regulation of transcription, DNA-templated
regulation of sodium-dependent phosphate transport
negative regulation of glucocorticoid receptor signaling pathway
transcription regulatory region sequence-specific DNA binding
transcription factor activity, transcription factor binding
DNA binding
damaged DNA binding
single-stranded DNA binding
deoxyribodipyrimidine photo-lyase activity
DNA (6-4) photolyase activity
protein binding
blue light photoreceptor activity
phosphatase binding
ubiquitin binding
FAD binding
MicroArray Probes:
Affymetrix
| Array Name | Probe IDs | Targets | Target Type |
|---|---|---|---|
| Affymetrix Human Transcriptome Array 2.0 | TC1100012994.hg.1 | ENST00000417225.6 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC1100012994.hg.1 | ENST00000417225.6 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC1100012994.hg.1 | ENST00000417225.6 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC1100012994.hg.1 | ENST00000417225.6 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC1100012994.hg.1 | ENST00000417225.6 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC1100012994.hg.1 | ENST00000417225.6 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC1100012994.hg.1 | ENST00000417225.6 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC1100012994.hg.1 | ENST00000417225.6 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC1100012994.hg.1 | ENST00000417225.6 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC1100012994.hg.1 | ENST00000417225.6 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC1100012994.hg.1 | ENST00000417225.6 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC1100012994.hg.1 | ENST00000417225.6 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC1100012994.hg.1 | ENST00000417225.6 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC1100012994.hg.1 | ENST00000417225.6 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC1100012994.hg.1 | ENST00000417225.6 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC1100012994.hg.1 | ENST00000417225.6 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC1100012994.hg.1 | ENST00000417225.6 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC1100012994.hg.1 | ENST00000417225.6 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC1100012994.hg.1 | ENST00000417225.6 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC1100012994.hg.1 | ENST00000417225.6 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC1100012994.hg.1 | ENST00000417225.6 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC1100012994.hg.1 | ENST00000443527.6 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC1100012994.hg.1 | ENST00000443527.6 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC1100012994.hg.1 | ENST00000443527.6 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC1100012994.hg.1 | ENST00000443527.6 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC1100012994.hg.1 | ENST00000443527.6 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC1100012994.hg.1 | ENST00000443527.6 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC1100012994.hg.1 | ENST00000443527.6 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC1100012994.hg.1 | ENST00000443527.6 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC1100012994.hg.1 | ENST00000443527.6 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC1100012994.hg.1 | ENST00000443527.6 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC1100012994.hg.1 | ENST00000443527.6 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC1100012994.hg.1 | ENST00000443527.6 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC1100012994.hg.1 | ENST00000443527.6 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC1100012994.hg.1 | ENST00000443527.6 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC1100012994.hg.1 | ENST00000443527.6 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC1100012994.hg.1 | ENST00000443527.6 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC1100012994.hg.1 | ENST00000443527.6 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC1100012994.hg.1 | ENST00000443527.6 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC1100012994.hg.1 | ENST00000443527.6 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC1100012994.hg.1 | ENST00000443527.6 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC1100012994.hg.1 | ENST00000443527.6 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC1100012994.hg.1 | ENST00000443527.6 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC1100012994.hg.1 | ENST00000473199.5 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC1100012994.hg.1 | ENST00000473199.5 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC1100012994.hg.1 | ENST00000473199.5 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC1100012994.hg.1 | ENST00000473199.5 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC1100012994.hg.1 | ENST00000488962.1 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC1100012994.hg.1 | ENST00000488962.1 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC1100012994.hg.1 | ENST00000488962.1 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC1100012994.hg.1 | ENST00000488962.1 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC1100012994.hg.1 | ENST00000495237.1 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC1100012994.hg.1 | ENST00000495237.1 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC1100012994.hg.1 | ENST00000495237.1 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC1100012994.hg.1 | ENST00000495237.1 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC1100012994.hg.1 | ENST00000495237.1 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC1100012994.hg.1 | ENST00000496571.5 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC1100012994.hg.1 | ENST00000496571.5 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC1100012994.hg.1 | ENST00000496571.5 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC1100012994.hg.1 | ENST00000496667.1 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC1100012994.hg.1 | ENST00000496667.1 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC1100012994.hg.1 | ENST00000496667.1 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC1100012994.hg.1 | ENST00000496667.1 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC1100012994.hg.1 | ENST00000525110.1 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC1100012994.hg.1 | ENST00000525110.1 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC1100012994.hg.1 | ENST00000525110.1 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC1100012994.hg.1 | ENST00000532390.5 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC1100012994.hg.1 | ENST00000532390.5 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC1100012994.hg.1 | ENST00000532390.5 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC1100012994.hg.1 | ENST00000532390.5 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC1100012994.hg.1 | ENST00000533779.1 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC1100012994.hg.1 | ENST00000533779.1 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC1100012994.hg.1 | ENST00000533779.1 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC1100012994.hg.1 | ENST00000533779.1 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC1100012994.hg.1 | ENST00000533779.1 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC1100012994.hg.1 | ENST00000533779.1 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC1100012994.hg.1 | ENST00000533779.1 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC1100012994.hg.1 | ENST00000533779.1 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC1100012994.hg.1 | ENST00000616080.1 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC1100012994.hg.1 | ENST00000616080.1 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC1100012994.hg.1 | ENST00000616080.1 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC1100012994.hg.1 | ENST00000616080.1 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC1100012994.hg.1 | ENST00000616080.1 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC1100012994.hg.1 | ENST00000616080.1 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC1100012994.hg.1 | ENST00000616080.1 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC1100012994.hg.1 | ENST00000616080.1 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC1100012994.hg.1 | ENST00000616080.1 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC1100012994.hg.1 | ENST00000616080.1 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC1100012994.hg.1 | ENST00000616080.1 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC1100012994.hg.1 | ENST00000616080.1 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC1100012994.hg.1 | ENST00000616080.1 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC1100012994.hg.1 | ENST00000616080.1 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC1100012994.hg.1 | ENST00000616080.1 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC1100012994.hg.1 | ENST00000616080.1 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC1100012994.hg.1 | ENST00000616080.1 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC1100012994.hg.1 | ENST00000616080.1 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC1100012994.hg.1 | ENST00000616080.1 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC1100012994.hg.1 | ENST00000616623.4 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC1100012994.hg.1 | ENST00000616623.4 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC1100012994.hg.1 | ENST00000616623.4 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC1100012994.hg.1 | ENST00000616623.4 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC1100012994.hg.1 | ENST00000616623.4 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC1100012994.hg.1 | ENST00000616623.4 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC1100012994.hg.1 | ENST00000616623.4 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC1100012994.hg.1 | ENST00000616623.4 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC1100012994.hg.1 | ENST00000616623.4 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC1100012994.hg.1 | ENST00000616623.4 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC1100012994.hg.1 | ENST00000616623.4 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC1100012994.hg.1 | ENST00000616623.4 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC1100012994.hg.1 | ENST00000616623.4 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC1100012994.hg.1 | ENST00000616623.4 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC1100012994.hg.1 | ENST00000616623.4 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC1100012994.hg.1 | ENST00000616623.4 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC1100012994.hg.1 | ENST00000616623.4 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC1100012994.hg.1 | ENST00000616623.4 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC1100012994.hg.1 | ENST00000616623.4 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC1100012994.hg.1 | ENST00000616623.4 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC1100012994.hg.1 | ENST00000616623.4 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC1100012994.hg.1 | ENST00000616623.4 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC1100012994.hg.1 | NM_001127457.2 | Refseq |
| Affymetrix Human Transcriptome Array 2.0 | TC1100012994.hg.1 | NM_001127457.2 | Refseq |
| Affymetrix Human Transcriptome Array 2.0 | TC1100012994.hg.1 | NM_001127457.2 | Refseq |
| Affymetrix Human Transcriptome Array 2.0 | TC1100012994.hg.1 | NM_001127457.2 | Refseq |
| Affymetrix Human Transcriptome Array 2.0 | TC1100012994.hg.1 | NM_001127457.2 | Refseq |
| Affymetrix Human Transcriptome Array 2.0 | TC1100012994.hg.1 | NM_001127457.2 | Refseq |
| Affymetrix Human Transcriptome Array 2.0 | TC1100012994.hg.1 | NM_001127457.2 | Refseq |
| Affymetrix Human Transcriptome Array 2.0 | TC1100012994.hg.1 | NM_001127457.2 | Refseq |
| Affymetrix Human Transcriptome Array 2.0 | TC1100012994.hg.1 | NM_001127457.2 | Refseq |
| Affymetrix Human Transcriptome Array 2.0 | TC1100012994.hg.1 | NM_001127457.2 | Refseq |
| Affymetrix Human Transcriptome Array 2.0 | TC1100012994.hg.1 | NM_001127457.2 | Refseq |
| Affymetrix Human Transcriptome Array 2.0 | TC1100012994.hg.1 | NM_001127457.2 | Refseq |
| Affymetrix Human Transcriptome Array 2.0 | TC1100012994.hg.1 | NM_001127457.2 | Refseq |
| Affymetrix Human Transcriptome Array 2.0 | TC1100012994.hg.1 | NM_001127457.2 | Refseq |
| Affymetrix Human Transcriptome Array 2.0 | TC1100012994.hg.1 | NM_001127457.2 | Refseq |
| Affymetrix Human Transcriptome Array 2.0 | TC1100012994.hg.1 | NM_001127457.2 | Refseq |
| Affymetrix Human Transcriptome Array 2.0 | TC1100012994.hg.1 | NM_001127457.2 | Refseq |
| Affymetrix Human Transcriptome Array 2.0 | TC1100012994.hg.1 | NM_001127457.2 | Refseq |
| Affymetrix Human Transcriptome Array 2.0 | TC1100012994.hg.1 | NM_001127457.2 | Refseq |
| Affymetrix Human Transcriptome Array 2.0 | TC1100012994.hg.1 | NM_001127457.2 | Refseq |
| Affymetrix Human Transcriptome Array 2.0 | TC1100012994.hg.1 | NM_001127457.2 | Refseq |
| Affymetrix Human Transcriptome Array 2.0 | TC1100012994.hg.1 | NM_001127457.2 | Refseq |
| Affymetrix Human Transcriptome Array 2.0 | TC1100012994.hg.1 | NM_021117.3 | Refseq |
| Affymetrix Human Transcriptome Array 2.0 | TC1100012994.hg.1 | NM_021117.3 | Refseq |
| Affymetrix Human Transcriptome Array 2.0 | TC1100012994.hg.1 | NM_021117.3 | Refseq |
| Affymetrix Human Transcriptome Array 2.0 | TC1100012994.hg.1 | NM_021117.3 | Refseq |
| Affymetrix Human Transcriptome Array 2.0 | TC1100012994.hg.1 | NM_021117.3 | Refseq |
| Affymetrix Human Transcriptome Array 2.0 | TC1100012994.hg.1 | NM_021117.3 | Refseq |
| Affymetrix Human Transcriptome Array 2.0 | TC1100012994.hg.1 | NM_021117.3 | Refseq |
| Affymetrix Human Transcriptome Array 2.0 | TC1100012994.hg.1 | NM_021117.3 | Refseq |
| Affymetrix Human Transcriptome Array 2.0 | TC1100012994.hg.1 | NM_021117.3 | Refseq |
| Affymetrix Human Transcriptome Array 2.0 | TC1100012994.hg.1 | NM_021117.3 | Refseq |
| Affymetrix Human Transcriptome Array 2.0 | TC1100012994.hg.1 | NM_021117.3 | Refseq |
| Affymetrix Human Transcriptome Array 2.0 | TC1100012994.hg.1 | NM_021117.3 | Refseq |
| Affymetrix Human Transcriptome Array 2.0 | TC1100012994.hg.1 | NM_021117.3 | Refseq |
| Affymetrix Human Transcriptome Array 2.0 | TC1100012994.hg.1 | NM_021117.3 | Refseq |
| Affymetrix Human Transcriptome Array 2.0 | TC1100012994.hg.1 | NM_021117.3 | Refseq |
| Affymetrix Human Transcriptome Array 2.0 | TC1100012994.hg.1 | NM_021117.3 | Refseq |
| Affymetrix Human Transcriptome Array 2.0 | TC1100012994.hg.1 | NM_021117.3 | Refseq |
| Affymetrix Human Transcriptome Array 2.0 | TC1100012994.hg.1 | NM_021117.3 | Refseq |
| Affymetrix Human Transcriptome Array 2.0 | TC1100012994.hg.1 | NM_021117.3 | Refseq |
| Affymetrix Human Transcriptome Array 2.0 | TC1100012994.hg.1 | NM_021117.3 | Refseq |
| Affymetrix Human Transcriptome Array 2.0 | TC1100012994.hg.1 | NM_021117.3 | Refseq |
| Affymetrix Human Transcriptome Array 2.0 | TC1100012994.hg.1 | NM_021117.3 | Refseq |

CRY2
Genomic Map