| Array Name |
Probe IDs |
Targets |
Target Type |
| Affymetrix Human Transcriptome Array 2.0 |
PSR10016920.hg.1 |
ENST00000277817.6 |
Ensembl |
| Affymetrix Human Transcriptome Array 2.0 |
PSR10016920.hg.1 |
ENST00000373700.4 |
Ensembl |
| Affymetrix Human Transcriptome Array 2.0 |
PSR10016920.hg.1 |
ENST00000395187.2 |
Ensembl |
| Affymetrix Human Transcriptome Array 2.0 |
PSR10016920.hg.1 |
ENST00000395198.3 |
Ensembl |
| Affymetrix Human Transcriptome Array 2.0 |
PSR10016920.hg.1 |
ENST00000412272.2 |
Ensembl |
| Affymetrix Human Transcriptome Array 2.0 |
PSR10016920.hg.1 |
NM_015601.3 |
Refseq |
| Affymetrix Human Transcriptome Array 2.0 |
PSR10016920.hg.1 |
NM_022079.2 |
Refseq |
| Affymetrix Human Transcriptome Array 2.0 |
PSR10016920.hg.1 |
TC10001341.hg.1 |
HTA TC |
| Affymetrix Human Clariom D Assay |
PSR1000173793.hg.1 |
ENST00000277817.10 |
Ensembl |
| Affymetrix Human Clariom D Assay |
PSR1000173793.hg.1 |
ENST00000373700.8 |
Ensembl |
| Affymetrix Human Clariom D Assay |
PSR1000173793.hg.1 |
ENST00000395198.7 |
Ensembl |
| Affymetrix Human Clariom D Assay |
PSR1000173793.hg.1 |
ENST00000412272.6 |
Ensembl |
| Affymetrix Human Clariom D Assay |
PSR1000173793.hg.1 |
ENST00000427635.6 |
Ensembl |
| Affymetrix Human Clariom D Assay |
PSR1000173793.hg.1 |
ENST00000473533.6 |
Ensembl |
| Affymetrix Human Clariom D Assay |
PSR1000173793.hg.1 |
NM_001278185.1 |
Refseq |
| Affymetrix Human Clariom D Assay |
PSR1000173793.hg.1 |
NM_001278186.1 |
Refseq |
| Affymetrix Human Clariom D Assay |
PSR1000173793.hg.1 |
NM_015601.3 |
Refseq |
| Affymetrix Human Clariom D Assay |
PSR1000173793.hg.1 |
NM_022079.2 |
Refseq |
| Affymetrix Human Clariom D Assay |
PSR1000173793.hg.1 |
ENST00000277817.10 |
Ensembl |
| Affymetrix Human Clariom D Assay |
PSR1000173793.hg.1 |
ENST00000373700.8 |
Ensembl |
| Affymetrix Human Clariom D Assay |
PSR1000173793.hg.1 |
ENST00000395198.7 |
Ensembl |
| Affymetrix Human Clariom D Assay |
PSR1000173793.hg.1 |
ENST00000412272.6 |
Ensembl |
| Affymetrix Human Clariom D Assay |
PSR1000173793.hg.1 |
ENST00000427635.6 |
Ensembl |
| Affymetrix Human Clariom D Assay |
PSR1000173793.hg.1 |
ENST00000473533.6 |
Ensembl |
| Affymetrix Human Clariom D Assay |
PSR1000173793.hg.1 |
NM_001278185.1 |
Refseq |
| Affymetrix Human Clariom D Assay |
PSR1000173793.hg.1 |
NM_001278186.1 |
Refseq |
| Affymetrix Human Clariom D Assay |
PSR1000173793.hg.1 |
NM_015601.3 |
Refseq |
| Affymetrix Human Clariom D Assay |
PSR1000173793.hg.1 |
NM_022079.2 |
Refseq |
| Affymetrix Human Clariom D Assay |
PSR1000173793.hg.1 |
TC1000010831.hg.1 |
Clariom D TC |