Assay Details
| Gene Symbol: PPP1R37 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | NM_019121.1 | NP_061994.1 | 2 - 3 | 453 | 56 | |
| XM_006723159.2 | XP_006723222.1 | 2 - 3 | 453 | 56 | ||
| GenBank mRNA | BC035704.1 | - | 2 - 3 | 675 | 56 | |
More Information
Updated Mapping Information
This assay targets the same location as HTA transcript cluster(s) TC19000642.hg.1
Set Membership: |
|
Gene Ontology Categories:
MicroArray Probes:
Affymetrix
| Array Name | Probe IDs | Targets | Target Type |
|---|---|---|---|
| Affymetrix Human Transcriptome Array 2.0 | PSR19008962.hg.1 | ENST00000221462.3 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | PSR19008962.hg.1 | ENST00000421905.1 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | PSR19008962.hg.1 | NM_019121.1 | Refseq |
| Affymetrix Human Transcriptome Array 2.0 | PSR19008962.hg.1 | TC19000642.hg.1 | HTA TC |
| Affymetrix Human Transcriptome Array 2.0 | TC1900011765.hg.1 | ENST00000221462.8 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC1900011765.hg.1 | ENST00000221462.8 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC1900011765.hg.1 | ENST00000221462.8 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC1900011765.hg.1 | ENST00000221462.8 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC1900011765.hg.1 | ENST00000221462.8 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC1900011765.hg.1 | ENST00000221462.8 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC1900011765.hg.1 | ENST00000221462.8 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC1900011765.hg.1 | ENST00000221462.8 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC1900011765.hg.1 | ENST00000221462.8 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC1900011765.hg.1 | ENST00000221462.8 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC1900011765.hg.1 | ENST00000221462.8 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC1900011765.hg.1 | ENST00000221462.8 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC1900011765.hg.1 | ENST00000221462.8 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC1900011765.hg.1 | ENST00000221462.8 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC1900011765.hg.1 | ENST00000221462.8 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC1900011765.hg.1 | ENST00000221462.8 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC1900011765.hg.1 | ENST00000221462.8 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC1900011765.hg.1 | ENST00000221462.8 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC1900011765.hg.1 | ENST00000221462.8 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC1900011765.hg.1 | ENST00000422370.2 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC1900011765.hg.1 | ENST00000422370.2 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC1900011765.hg.1 | ENST00000422370.2 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC1900011765.hg.1 | ENST00000422370.2 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC1900011765.hg.1 | ENST00000422370.2 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC1900011765.hg.1 | ENST00000422370.2 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC1900011765.hg.1 | ENST00000422370.2 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC1900011765.hg.1 | ENST00000422370.2 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC1900011765.hg.1 | ENST00000422370.2 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC1900011765.hg.1 | ENST00000422370.2 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC1900011765.hg.1 | ENST00000422370.2 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC1900011765.hg.1 | ENST00000422370.2 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC1900011765.hg.1 | ENST00000422370.2 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC1900011765.hg.1 | ENST00000422370.2 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC1900011765.hg.1 | ENST00000422370.2 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC1900011765.hg.1 | ENST00000422370.2 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC1900011765.hg.1 | ENST00000496125.2 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC1900011765.hg.1 | ENST00000496125.2 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC1900011765.hg.1 | ENST00000496125.2 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC1900011765.hg.1 | ENST00000540059.1 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC1900011765.hg.1 | ENST00000540059.1 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC1900011765.hg.1 | ENST00000540059.1 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC1900011765.hg.1 | ENST00000540059.1 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC1900011765.hg.1 | ENST00000540059.1 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC1900011765.hg.1 | ENST00000544069.2 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC1900011765.hg.1 | ENST00000544069.2 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC1900011765.hg.1 | ENST00000544069.2 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC1900011765.hg.1 | ENST00000544069.2 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC1900011765.hg.1 | ENST00000544069.2 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC1900011765.hg.1 | ENST00000544069.2 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC1900011765.hg.1 | ENST00000544069.2 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC1900011765.hg.1 | ENST00000544897.5 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC1900011765.hg.1 | ENST00000544897.5 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC1900011765.hg.1 | ENST00000544897.5 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC1900011765.hg.1 | ENST00000544897.5 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC1900011765.hg.1 | ENST00000544897.5 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC1900011765.hg.1 | ENST00000544897.5 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC1900011765.hg.1 | ENST00000545910.1 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC1900011765.hg.1 | ENST00000545910.1 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC1900011765.hg.1 | ENST00000545910.1 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC1900011765.hg.1 | ENST00000545910.1 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC1900011765.hg.1 | ENST00000545910.1 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC1900011765.hg.1 | NM_019121.1 | Refseq |
| Affymetrix Human Transcriptome Array 2.0 | TC1900011765.hg.1 | NM_019121.1 | Refseq |
| Affymetrix Human Transcriptome Array 2.0 | TC1900011765.hg.1 | NM_019121.1 | Refseq |
| Affymetrix Human Transcriptome Array 2.0 | TC1900011765.hg.1 | NM_019121.1 | Refseq |
| Affymetrix Human Transcriptome Array 2.0 | TC1900011765.hg.1 | NM_019121.1 | Refseq |
| Affymetrix Human Transcriptome Array 2.0 | TC1900011765.hg.1 | NM_019121.1 | Refseq |
| Affymetrix Human Transcriptome Array 2.0 | TC1900011765.hg.1 | NM_019121.1 | Refseq |
| Affymetrix Human Transcriptome Array 2.0 | TC1900011765.hg.1 | NM_019121.1 | Refseq |
| Affymetrix Human Transcriptome Array 2.0 | TC1900011765.hg.1 | NM_019121.1 | Refseq |
| Affymetrix Human Transcriptome Array 2.0 | TC1900011765.hg.1 | NM_019121.1 | Refseq |
| Affymetrix Human Transcriptome Array 2.0 | TC1900011765.hg.1 | NM_019121.1 | Refseq |
| Affymetrix Human Transcriptome Array 2.0 | TC1900011765.hg.1 | NM_019121.1 | Refseq |
| Affymetrix Human Transcriptome Array 2.0 | TC1900011765.hg.1 | NM_019121.1 | Refseq |
| Affymetrix Human Transcriptome Array 2.0 | TC1900011765.hg.1 | NM_019121.1 | Refseq |
| Affymetrix Human Transcriptome Array 2.0 | TC1900011765.hg.1 | NM_019121.1 | Refseq |
| Affymetrix Human Transcriptome Array 2.0 | TC1900011765.hg.1 | NM_019121.1 | Refseq |
| Affymetrix Human Transcriptome Array 2.0 | TC1900011765.hg.1 | NM_019121.1 | Refseq |
| Affymetrix Human Transcriptome Array 2.0 | TC1900011765.hg.1 | NM_019121.1 | Refseq |
| Affymetrix Human Clariom D Assay | PSR1900201659.hg.1 | ENST00000221462.8 | Ensembl |
| Affymetrix Human Clariom D Assay | PSR1900201659.hg.1 | ENST00000544897.5 | Ensembl |
| Affymetrix Human Clariom D Assay | PSR1900201659.hg.1 | ENST00000545910.1 | Ensembl |
| Affymetrix Human Clariom D Assay | PSR1900201659.hg.1 | NM_019121.1 | Refseq |
| Affymetrix Human Clariom D Assay | PSR1900201659.hg.1 | TC1900011765.hg.1 | Clariom D TC |
| Affymetrix Human Clariom D Assay | PSR1900201659.hg.1 | ENST00000221462.8 | Ensembl |
| Affymetrix Human Clariom D Assay | PSR1900201659.hg.1 | ENST00000544897.5 | Ensembl |
| Affymetrix Human Clariom D Assay | PSR1900201659.hg.1 | ENST00000545910.1 | Ensembl |
| Affymetrix Human Clariom D Assay | PSR1900201659.hg.1 | NM_019121.1 | Refseq |
| Affymetrix Human Clariom D Assay | PSR1900201660.hg.1 | ENST00000221462.8 | Ensembl |
| Affymetrix Human Clariom D Assay | PSR1900201660.hg.1 | ENST00000544897.5 | Ensembl |
| Affymetrix Human Clariom D Assay | PSR1900201660.hg.1 | ENST00000545910.1 | Ensembl |
| Affymetrix Human Clariom D Assay | PSR1900201660.hg.1 | NM_019121.1 | Refseq |
| Affymetrix Human Clariom D Assay | PSR1900201660.hg.1 | TC1900011765.hg.1 | Clariom D TC |
| Affymetrix Human Clariom D Assay | PSR1900201660.hg.1 | ENST00000221462.8 | Ensembl |
| Affymetrix Human Clariom D Assay | PSR1900201660.hg.1 | ENST00000544897.5 | Ensembl |
| Affymetrix Human Clariom D Assay | PSR1900201660.hg.1 | ENST00000545910.1 | Ensembl |
| Affymetrix Human Clariom D Assay | PSR1900201660.hg.1 | NM_019121.1 | Refseq |
| Affymetrix Human Clariom D Assay | TC1900011765.hg.1 | ENST00000221462.8 | Ensembl |
| Affymetrix Human Clariom D Assay | TC1900011765.hg.1 | ENST00000221462.8 | Ensembl |
| Affymetrix Human Clariom D Assay | TC1900011765.hg.1 | ENST00000221462.8 | Ensembl |
| Affymetrix Human Clariom D Assay | TC1900011765.hg.1 | ENST00000221462.8 | Ensembl |
| Affymetrix Human Clariom D Assay | TC1900011765.hg.1 | ENST00000221462.8 | Ensembl |
| Affymetrix Human Clariom D Assay | TC1900011765.hg.1 | ENST00000221462.8 | Ensembl |
| Affymetrix Human Clariom D Assay | TC1900011765.hg.1 | ENST00000221462.8 | Ensembl |
| Affymetrix Human Clariom D Assay | TC1900011765.hg.1 | ENST00000221462.8 | Ensembl |
| Affymetrix Human Clariom D Assay | TC1900011765.hg.1 | ENST00000221462.8 | Ensembl |
| Affymetrix Human Clariom D Assay | TC1900011765.hg.1 | ENST00000221462.8 | Ensembl |
| Affymetrix Human Clariom D Assay | TC1900011765.hg.1 | ENST00000221462.8 | Ensembl |
| Affymetrix Human Clariom D Assay | TC1900011765.hg.1 | ENST00000221462.8 | Ensembl |
| Affymetrix Human Clariom D Assay | TC1900011765.hg.1 | ENST00000221462.8 | Ensembl |
| Affymetrix Human Clariom D Assay | TC1900011765.hg.1 | ENST00000221462.8 | Ensembl |
| Affymetrix Human Clariom D Assay | TC1900011765.hg.1 | ENST00000221462.8 | Ensembl |
| Affymetrix Human Clariom D Assay | TC1900011765.hg.1 | ENST00000221462.8 | Ensembl |
| Affymetrix Human Clariom D Assay | TC1900011765.hg.1 | ENST00000221462.8 | Ensembl |
| Affymetrix Human Clariom D Assay | TC1900011765.hg.1 | ENST00000221462.8 | Ensembl |
| Affymetrix Human Clariom D Assay | TC1900011765.hg.1 | ENST00000221462.8 | Ensembl |
| Affymetrix Human Clariom D Assay | TC1900011765.hg.1 | ENST00000422370.2 | Ensembl |
| Affymetrix Human Clariom D Assay | TC1900011765.hg.1 | ENST00000422370.2 | Ensembl |
| Affymetrix Human Clariom D Assay | TC1900011765.hg.1 | ENST00000422370.2 | Ensembl |
| Affymetrix Human Clariom D Assay | TC1900011765.hg.1 | ENST00000422370.2 | Ensembl |
| Affymetrix Human Clariom D Assay | TC1900011765.hg.1 | ENST00000422370.2 | Ensembl |
| Affymetrix Human Clariom D Assay | TC1900011765.hg.1 | ENST00000422370.2 | Ensembl |
| Affymetrix Human Clariom D Assay | TC1900011765.hg.1 | ENST00000422370.2 | Ensembl |
| Affymetrix Human Clariom D Assay | TC1900011765.hg.1 | ENST00000422370.2 | Ensembl |
| Affymetrix Human Clariom D Assay | TC1900011765.hg.1 | ENST00000422370.2 | Ensembl |
| Affymetrix Human Clariom D Assay | TC1900011765.hg.1 | ENST00000422370.2 | Ensembl |
| Affymetrix Human Clariom D Assay | TC1900011765.hg.1 | ENST00000422370.2 | Ensembl |
| Affymetrix Human Clariom D Assay | TC1900011765.hg.1 | ENST00000422370.2 | Ensembl |
| Affymetrix Human Clariom D Assay | TC1900011765.hg.1 | ENST00000422370.2 | Ensembl |
| Affymetrix Human Clariom D Assay | TC1900011765.hg.1 | ENST00000422370.2 | Ensembl |
| Affymetrix Human Clariom D Assay | TC1900011765.hg.1 | ENST00000422370.2 | Ensembl |
| Affymetrix Human Clariom D Assay | TC1900011765.hg.1 | ENST00000422370.2 | Ensembl |
| Affymetrix Human Clariom D Assay | TC1900011765.hg.1 | ENST00000496125.2 | Ensembl |
| Affymetrix Human Clariom D Assay | TC1900011765.hg.1 | ENST00000496125.2 | Ensembl |
| Affymetrix Human Clariom D Assay | TC1900011765.hg.1 | ENST00000496125.2 | Ensembl |
| Affymetrix Human Clariom D Assay | TC1900011765.hg.1 | ENST00000540059.1 | Ensembl |
| Affymetrix Human Clariom D Assay | TC1900011765.hg.1 | ENST00000540059.1 | Ensembl |
| Affymetrix Human Clariom D Assay | TC1900011765.hg.1 | ENST00000540059.1 | Ensembl |
| Affymetrix Human Clariom D Assay | TC1900011765.hg.1 | ENST00000540059.1 | Ensembl |
| Affymetrix Human Clariom D Assay | TC1900011765.hg.1 | ENST00000540059.1 | Ensembl |
| Affymetrix Human Clariom D Assay | TC1900011765.hg.1 | ENST00000544069.2 | Ensembl |
| Affymetrix Human Clariom D Assay | TC1900011765.hg.1 | ENST00000544069.2 | Ensembl |
| Affymetrix Human Clariom D Assay | TC1900011765.hg.1 | ENST00000544069.2 | Ensembl |
| Affymetrix Human Clariom D Assay | TC1900011765.hg.1 | ENST00000544069.2 | Ensembl |
| Affymetrix Human Clariom D Assay | TC1900011765.hg.1 | ENST00000544069.2 | Ensembl |
| Affymetrix Human Clariom D Assay | TC1900011765.hg.1 | ENST00000544069.2 | Ensembl |
| Affymetrix Human Clariom D Assay | TC1900011765.hg.1 | ENST00000544069.2 | Ensembl |
| Affymetrix Human Clariom D Assay | TC1900011765.hg.1 | ENST00000544897.5 | Ensembl |
| Affymetrix Human Clariom D Assay | TC1900011765.hg.1 | ENST00000544897.5 | Ensembl |
| Affymetrix Human Clariom D Assay | TC1900011765.hg.1 | ENST00000544897.5 | Ensembl |
| Affymetrix Human Clariom D Assay | TC1900011765.hg.1 | ENST00000544897.5 | Ensembl |
| Affymetrix Human Clariom D Assay | TC1900011765.hg.1 | ENST00000544897.5 | Ensembl |
| Affymetrix Human Clariom D Assay | TC1900011765.hg.1 | ENST00000544897.5 | Ensembl |
| Affymetrix Human Clariom D Assay | TC1900011765.hg.1 | ENST00000545910.1 | Ensembl |
| Affymetrix Human Clariom D Assay | TC1900011765.hg.1 | ENST00000545910.1 | Ensembl |
| Affymetrix Human Clariom D Assay | TC1900011765.hg.1 | ENST00000545910.1 | Ensembl |
| Affymetrix Human Clariom D Assay | TC1900011765.hg.1 | ENST00000545910.1 | Ensembl |
| Affymetrix Human Clariom D Assay | TC1900011765.hg.1 | ENST00000545910.1 | Ensembl |
| Affymetrix Human Clariom D Assay | TC1900011765.hg.1 | NM_019121.1 | Refseq |
| Affymetrix Human Clariom D Assay | TC1900011765.hg.1 | NM_019121.1 | Refseq |
| Affymetrix Human Clariom D Assay | TC1900011765.hg.1 | NM_019121.1 | Refseq |
| Affymetrix Human Clariom D Assay | TC1900011765.hg.1 | NM_019121.1 | Refseq |
| Affymetrix Human Clariom D Assay | TC1900011765.hg.1 | NM_019121.1 | Refseq |
| Affymetrix Human Clariom D Assay | TC1900011765.hg.1 | NM_019121.1 | Refseq |
| Affymetrix Human Clariom D Assay | TC1900011765.hg.1 | NM_019121.1 | Refseq |
| Affymetrix Human Clariom D Assay | TC1900011765.hg.1 | NM_019121.1 | Refseq |
| Affymetrix Human Clariom D Assay | TC1900011765.hg.1 | NM_019121.1 | Refseq |
| Affymetrix Human Clariom D Assay | TC1900011765.hg.1 | NM_019121.1 | Refseq |
| Affymetrix Human Clariom D Assay | TC1900011765.hg.1 | NM_019121.1 | Refseq |
| Affymetrix Human Clariom D Assay | TC1900011765.hg.1 | NM_019121.1 | Refseq |
| Affymetrix Human Clariom D Assay | TC1900011765.hg.1 | NM_019121.1 | Refseq |
| Affymetrix Human Clariom D Assay | TC1900011765.hg.1 | NM_019121.1 | Refseq |
| Affymetrix Human Clariom D Assay | TC1900011765.hg.1 | NM_019121.1 | Refseq |
| Affymetrix Human Clariom D Assay | TC1900011765.hg.1 | NM_019121.1 | Refseq |
| Affymetrix Human Clariom D Assay | TC1900011765.hg.1 | NM_019121.1 | Refseq |
| Affymetrix Human Clariom D Assay | TC1900011765.hg.1 | NM_019121.1 | Refseq |

PPP1R37
Genomic Map