Assay Details
Gene Symbol: GPD1L | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | NM_015141.3 | NP_055956.1 | 3 - 4 | 572 | 121 | |
GenBank mRNA | AK292808.1 | - | 3 - 4 | 405 | 121 | |
BC006168.1 | - | 1 - 2 | 96 | 121 | ||
BC028726.2 | - | 3 - 4 | 431 | 4820730 | 121 | |
D42047.1 | - | 3 - 4 | 552 | 121 |

More Information
Updated Mapping Information

This assay targets the same location as HTA transcript cluster(s) TC03000157.hg.1
Set Membership: |
![]() ![]() ![]() ![]() ![]() |
Matching Ambion siRNAs:
Panther Classification:
Gene Ontology Categories:
Function(s)
Process(es)
regulation of heart rate
carbohydrate metabolic process
phosphatidic acid biosynthetic process
NADH metabolic process
positive regulation of sodium ion transport
triglyceride biosynthetic process
NAD metabolic process
negative regulation of peptidyl-serine phosphorylation
glycerol-3-phosphate catabolic process
oxidation-reduction process
regulation of ventricular cardiac muscle cell membrane depolarization
ventricular cardiac muscle cell action potential
negative regulation of protein kinase C signaling
positive regulation of protein localization to cell surface
regulation of sodium ion transmembrane transporter activity
glycerol-3-phosphate dehydrogenase [NAD+] activity
sodium channel regulator activity
protein homodimerization activity
ion channel binding
NAD binding
regulation of heart rate
carbohydrate metabolic process
phosphatidic acid biosynthetic process
NADH metabolic process
positive regulation of sodium ion transport
triglyceride biosynthetic process
NAD metabolic process
negative regulation of peptidyl-serine phosphorylation
glycerol-3-phosphate catabolic process
oxidation-reduction process
regulation of ventricular cardiac muscle cell membrane depolarization
ventricular cardiac muscle cell action potential
negative regulation of protein kinase C signaling
positive regulation of protein localization to cell surface
regulation of sodium ion transmembrane transporter activity
glycerol-3-phosphate dehydrogenase [NAD+] activity
sodium channel regulator activity
protein homodimerization activity
ion channel binding
NAD binding
MicroArray Probes:
Affymetrix
Array Name | Probe IDs | Targets | Target Type |
---|---|---|---|
Affymetrix Human Transcriptome Array 2.0 | PSR03002800.hg.1 | TC03000157.hg.1 | HTA TC |
Affymetrix Human Transcriptome Array 2.0 | PSR03002800.hg.1 | ENST00000282541.5 | Ensembl |
Affymetrix Human Transcriptome Array 2.0 | PSR03002800.hg.1 | NM_015141.3 | Refseq |
Affymetrix Human Transcriptome Array 2.0 | TC0300006956.hg.1 | ENST00000282541.9 | Ensembl |
Affymetrix Human Transcriptome Array 2.0 | TC0300006956.hg.1 | ENST00000282541.9 | Ensembl |
Affymetrix Human Transcriptome Array 2.0 | TC0300006956.hg.1 | ENST00000282541.9 | Ensembl |
Affymetrix Human Transcriptome Array 2.0 | TC0300006956.hg.1 | ENST00000282541.9 | Ensembl |
Affymetrix Human Transcriptome Array 2.0 | TC0300006956.hg.1 | ENST00000282541.9 | Ensembl |
Affymetrix Human Transcriptome Array 2.0 | TC0300006956.hg.1 | ENST00000282541.9 | Ensembl |
Affymetrix Human Transcriptome Array 2.0 | TC0300006956.hg.1 | ENST00000282541.9 | Ensembl |
Affymetrix Human Transcriptome Array 2.0 | TC0300006956.hg.1 | ENST00000282541.9 | Ensembl |
Affymetrix Human Transcriptome Array 2.0 | TC0300006956.hg.1 | ENST00000282541.9 | Ensembl |
Affymetrix Human Transcriptome Array 2.0 | TC0300006956.hg.1 | ENST00000282541.9 | Ensembl |
Affymetrix Human Transcriptome Array 2.0 | TC0300006956.hg.1 | ENST00000282541.9 | Ensembl |
Affymetrix Human Transcriptome Array 2.0 | TC0300006956.hg.1 | ENST00000282541.9 | Ensembl |
Affymetrix Human Transcriptome Array 2.0 | TC0300006956.hg.1 | ENST00000282541.9 | Ensembl |
Affymetrix Human Transcriptome Array 2.0 | TC0300006956.hg.1 | ENST00000282541.9 | Ensembl |
Affymetrix Human Transcriptome Array 2.0 | TC0300006956.hg.1 | ENST00000425459.5 | Ensembl |
Affymetrix Human Transcriptome Array 2.0 | TC0300006956.hg.1 | ENST00000425459.5 | Ensembl |
Affymetrix Human Transcriptome Array 2.0 | TC0300006956.hg.1 | ENST00000425459.5 | Ensembl |
Affymetrix Human Transcriptome Array 2.0 | TC0300006956.hg.1 | ENST00000425459.5 | Ensembl |
Affymetrix Human Transcriptome Array 2.0 | TC0300006956.hg.1 | ENST00000425459.5 | Ensembl |
Affymetrix Human Transcriptome Array 2.0 | TC0300006956.hg.1 | ENST00000425459.5 | Ensembl |
Affymetrix Human Transcriptome Array 2.0 | TC0300006956.hg.1 | ENST00000425459.5 | Ensembl |
Affymetrix Human Transcriptome Array 2.0 | TC0300006956.hg.1 | ENST00000428684.1 | Ensembl |
Affymetrix Human Transcriptome Array 2.0 | TC0300006956.hg.1 | ENST00000428684.1 | Ensembl |
Affymetrix Human Transcriptome Array 2.0 | TC0300006956.hg.1 | ENST00000428684.1 | Ensembl |
Affymetrix Human Transcriptome Array 2.0 | TC0300006956.hg.1 | ENST00000428684.1 | Ensembl |
Affymetrix Human Transcriptome Array 2.0 | TC0300006956.hg.1 | ENST00000428684.1 | Ensembl |
Affymetrix Human Transcriptome Array 2.0 | TC0300006956.hg.1 | ENST00000428684.1 | Ensembl |
Affymetrix Human Transcriptome Array 2.0 | TC0300006956.hg.1 | ENST00000428684.1 | Ensembl |
Affymetrix Human Transcriptome Array 2.0 | TC0300006956.hg.1 | ENST00000428684.1 | Ensembl |
Affymetrix Human Transcriptome Array 2.0 | TC0300006956.hg.1 | ENST00000428684.1 | Ensembl |
Affymetrix Human Transcriptome Array 2.0 | TC0300006956.hg.1 | ENST00000429432.5 | Ensembl |
Affymetrix Human Transcriptome Array 2.0 | TC0300006956.hg.1 | ENST00000429432.5 | Ensembl |
Affymetrix Human Transcriptome Array 2.0 | TC0300006956.hg.1 | ENST00000429432.5 | Ensembl |
Affymetrix Human Transcriptome Array 2.0 | TC0300006956.hg.1 | ENST00000429432.5 | Ensembl |
Affymetrix Human Transcriptome Array 2.0 | TC0300006956.hg.1 | ENST00000431009.1 | Ensembl |
Affymetrix Human Transcriptome Array 2.0 | TC0300006956.hg.1 | ENST00000431009.1 | Ensembl |
Affymetrix Human Transcriptome Array 2.0 | TC0300006956.hg.1 | ENST00000431009.1 | Ensembl |
Affymetrix Human Transcriptome Array 2.0 | TC0300006956.hg.1 | ENST00000431009.1 | Ensembl |
Affymetrix Human Transcriptome Array 2.0 | TC0300006956.hg.1 | ENST00000431009.1 | Ensembl |
Affymetrix Human Transcriptome Array 2.0 | TC0300006956.hg.1 | ENST00000431009.1 | Ensembl |
Affymetrix Human Transcriptome Array 2.0 | TC0300006956.hg.1 | ENST00000474846.5 | Ensembl |
Affymetrix Human Transcriptome Array 2.0 | TC0300006956.hg.1 | ENST00000474846.5 | Ensembl |
Affymetrix Human Transcriptome Array 2.0 | TC0300006956.hg.1 | ENST00000474846.5 | Ensembl |
Affymetrix Human Transcriptome Array 2.0 | TC0300006956.hg.1 | ENST00000474846.5 | Ensembl |
Affymetrix Human Transcriptome Array 2.0 | TC0300006956.hg.1 | ENST00000474846.5 | Ensembl |
Affymetrix Human Transcriptome Array 2.0 | TC0300006956.hg.1 | ENST00000474846.5 | Ensembl |
Affymetrix Human Transcriptome Array 2.0 | TC0300006956.hg.1 | ENST00000474846.5 | Ensembl |
Affymetrix Human Transcriptome Array 2.0 | TC0300006956.hg.1 | ENST00000474846.5 | Ensembl |
Affymetrix Human Transcriptome Array 2.0 | TC0300006956.hg.1 | ENST00000496151.1 | Ensembl |
Affymetrix Human Transcriptome Array 2.0 | TC0300006956.hg.1 | ENST00000496151.1 | Ensembl |
Affymetrix Human Transcriptome Array 2.0 | TC0300006956.hg.1 | ENST00000496151.1 | Ensembl |
Affymetrix Human Transcriptome Array 2.0 | TC0300006956.hg.1 | ENST00000496151.1 | Ensembl |
Affymetrix Human Transcriptome Array 2.0 | TC0300006956.hg.1 | ENST00000496151.1 | Ensembl |
Affymetrix Human Transcriptome Array 2.0 | TC0300006956.hg.1 | ENST00000496151.1 | Ensembl |
Affymetrix Human Transcriptome Array 2.0 | TC0300006956.hg.1 | NM_015141.3 | Refseq |
Affymetrix Human Transcriptome Array 2.0 | TC0300006956.hg.1 | NM_015141.3 | Refseq |
Affymetrix Human Transcriptome Array 2.0 | TC0300006956.hg.1 | NM_015141.3 | Refseq |
Affymetrix Human Transcriptome Array 2.0 | TC0300006956.hg.1 | NM_015141.3 | Refseq |
Affymetrix Human Transcriptome Array 2.0 | TC0300006956.hg.1 | NM_015141.3 | Refseq |
Affymetrix Human Transcriptome Array 2.0 | TC0300006956.hg.1 | NM_015141.3 | Refseq |
Affymetrix Human Transcriptome Array 2.0 | TC0300006956.hg.1 | NM_015141.3 | Refseq |
Affymetrix Human Transcriptome Array 2.0 | TC0300006956.hg.1 | NM_015141.3 | Refseq |
Affymetrix Human Transcriptome Array 2.0 | TC0300006956.hg.1 | NM_015141.3 | Refseq |
Affymetrix Human Transcriptome Array 2.0 | TC0300006956.hg.1 | NM_015141.3 | Refseq |
Affymetrix Human Transcriptome Array 2.0 | TC0300006956.hg.1 | NM_015141.3 | Refseq |
Affymetrix Human Transcriptome Array 2.0 | TC0300006956.hg.1 | NM_015141.3 | Refseq |
Affymetrix Human Transcriptome Array 2.0 | TC0300006956.hg.1 | NM_015141.3 | Refseq |
Affymetrix Human Transcriptome Array 2.0 | TC0300006956.hg.1 | NM_015141.3 | Refseq |
Affymetrix Human Clariom D Assay | TC0300006956.hg.1 | ENST00000282541.9 | Ensembl |
Affymetrix Human Clariom D Assay | TC0300006956.hg.1 | ENST00000282541.9 | Ensembl |
Affymetrix Human Clariom D Assay | TC0300006956.hg.1 | ENST00000282541.9 | Ensembl |
Affymetrix Human Clariom D Assay | TC0300006956.hg.1 | ENST00000282541.9 | Ensembl |
Affymetrix Human Clariom D Assay | TC0300006956.hg.1 | ENST00000282541.9 | Ensembl |
Affymetrix Human Clariom D Assay | TC0300006956.hg.1 | ENST00000282541.9 | Ensembl |
Affymetrix Human Clariom D Assay | TC0300006956.hg.1 | ENST00000282541.9 | Ensembl |
Affymetrix Human Clariom D Assay | TC0300006956.hg.1 | ENST00000282541.9 | Ensembl |
Affymetrix Human Clariom D Assay | TC0300006956.hg.1 | ENST00000282541.9 | Ensembl |
Affymetrix Human Clariom D Assay | TC0300006956.hg.1 | ENST00000282541.9 | Ensembl |
Affymetrix Human Clariom D Assay | TC0300006956.hg.1 | ENST00000282541.9 | Ensembl |
Affymetrix Human Clariom D Assay | TC0300006956.hg.1 | ENST00000282541.9 | Ensembl |
Affymetrix Human Clariom D Assay | TC0300006956.hg.1 | ENST00000282541.9 | Ensembl |
Affymetrix Human Clariom D Assay | TC0300006956.hg.1 | ENST00000282541.9 | Ensembl |
Affymetrix Human Clariom D Assay | TC0300006956.hg.1 | ENST00000425459.5 | Ensembl |
Affymetrix Human Clariom D Assay | TC0300006956.hg.1 | ENST00000425459.5 | Ensembl |
Affymetrix Human Clariom D Assay | TC0300006956.hg.1 | ENST00000425459.5 | Ensembl |
Affymetrix Human Clariom D Assay | TC0300006956.hg.1 | ENST00000425459.5 | Ensembl |
Affymetrix Human Clariom D Assay | TC0300006956.hg.1 | ENST00000425459.5 | Ensembl |
Affymetrix Human Clariom D Assay | TC0300006956.hg.1 | ENST00000425459.5 | Ensembl |
Affymetrix Human Clariom D Assay | TC0300006956.hg.1 | ENST00000425459.5 | Ensembl |
Affymetrix Human Clariom D Assay | TC0300006956.hg.1 | ENST00000428684.1 | Ensembl |
Affymetrix Human Clariom D Assay | TC0300006956.hg.1 | ENST00000428684.1 | Ensembl |
Affymetrix Human Clariom D Assay | TC0300006956.hg.1 | ENST00000428684.1 | Ensembl |
Affymetrix Human Clariom D Assay | TC0300006956.hg.1 | ENST00000428684.1 | Ensembl |
Affymetrix Human Clariom D Assay | TC0300006956.hg.1 | ENST00000428684.1 | Ensembl |
Affymetrix Human Clariom D Assay | TC0300006956.hg.1 | ENST00000428684.1 | Ensembl |
Affymetrix Human Clariom D Assay | TC0300006956.hg.1 | ENST00000428684.1 | Ensembl |
Affymetrix Human Clariom D Assay | TC0300006956.hg.1 | ENST00000428684.1 | Ensembl |
Affymetrix Human Clariom D Assay | TC0300006956.hg.1 | ENST00000428684.1 | Ensembl |
Affymetrix Human Clariom D Assay | TC0300006956.hg.1 | ENST00000429432.5 | Ensembl |
Affymetrix Human Clariom D Assay | TC0300006956.hg.1 | ENST00000429432.5 | Ensembl |
Affymetrix Human Clariom D Assay | TC0300006956.hg.1 | ENST00000429432.5 | Ensembl |
Affymetrix Human Clariom D Assay | TC0300006956.hg.1 | ENST00000429432.5 | Ensembl |
Affymetrix Human Clariom D Assay | TC0300006956.hg.1 | ENST00000431009.1 | Ensembl |
Affymetrix Human Clariom D Assay | TC0300006956.hg.1 | ENST00000431009.1 | Ensembl |
Affymetrix Human Clariom D Assay | TC0300006956.hg.1 | ENST00000431009.1 | Ensembl |
Affymetrix Human Clariom D Assay | TC0300006956.hg.1 | ENST00000431009.1 | Ensembl |
Affymetrix Human Clariom D Assay | TC0300006956.hg.1 | ENST00000431009.1 | Ensembl |
Affymetrix Human Clariom D Assay | TC0300006956.hg.1 | ENST00000431009.1 | Ensembl |
Affymetrix Human Clariom D Assay | TC0300006956.hg.1 | ENST00000474846.5 | Ensembl |
Affymetrix Human Clariom D Assay | TC0300006956.hg.1 | ENST00000474846.5 | Ensembl |
Affymetrix Human Clariom D Assay | TC0300006956.hg.1 | ENST00000474846.5 | Ensembl |
Affymetrix Human Clariom D Assay | TC0300006956.hg.1 | ENST00000474846.5 | Ensembl |
Affymetrix Human Clariom D Assay | TC0300006956.hg.1 | ENST00000474846.5 | Ensembl |
Affymetrix Human Clariom D Assay | TC0300006956.hg.1 | ENST00000474846.5 | Ensembl |
Affymetrix Human Clariom D Assay | TC0300006956.hg.1 | ENST00000474846.5 | Ensembl |
Affymetrix Human Clariom D Assay | TC0300006956.hg.1 | ENST00000474846.5 | Ensembl |
Affymetrix Human Clariom D Assay | TC0300006956.hg.1 | ENST00000496151.1 | Ensembl |
Affymetrix Human Clariom D Assay | TC0300006956.hg.1 | ENST00000496151.1 | Ensembl |
Affymetrix Human Clariom D Assay | TC0300006956.hg.1 | ENST00000496151.1 | Ensembl |
Affymetrix Human Clariom D Assay | TC0300006956.hg.1 | ENST00000496151.1 | Ensembl |
Affymetrix Human Clariom D Assay | TC0300006956.hg.1 | ENST00000496151.1 | Ensembl |
Affymetrix Human Clariom D Assay | TC0300006956.hg.1 | ENST00000496151.1 | Ensembl |
Affymetrix Human Clariom D Assay | TC0300006956.hg.1 | NM_015141.3 | Refseq |
Affymetrix Human Clariom D Assay | TC0300006956.hg.1 | NM_015141.3 | Refseq |
Affymetrix Human Clariom D Assay | TC0300006956.hg.1 | NM_015141.3 | Refseq |
Affymetrix Human Clariom D Assay | TC0300006956.hg.1 | NM_015141.3 | Refseq |
Affymetrix Human Clariom D Assay | TC0300006956.hg.1 | NM_015141.3 | Refseq |
Affymetrix Human Clariom D Assay | TC0300006956.hg.1 | NM_015141.3 | Refseq |
Affymetrix Human Clariom D Assay | TC0300006956.hg.1 | NM_015141.3 | Refseq |
Affymetrix Human Clariom D Assay | TC0300006956.hg.1 | NM_015141.3 | Refseq |
Affymetrix Human Clariom D Assay | TC0300006956.hg.1 | NM_015141.3 | Refseq |
Affymetrix Human Clariom D Assay | TC0300006956.hg.1 | NM_015141.3 | Refseq |
Affymetrix Human Clariom D Assay | TC0300006956.hg.1 | NM_015141.3 | Refseq |
Affymetrix Human Clariom D Assay | TC0300006956.hg.1 | NM_015141.3 | Refseq |
Affymetrix Human Clariom D Assay | TC0300006956.hg.1 | NM_015141.3 | Refseq |
Affymetrix Human Clariom D Assay | TC0300006956.hg.1 | NM_015141.3 | Refseq |
Affymetrix Human Clariom D Assay | PSR0300148412.hg.1 | ENST00000282541.9 | Ensembl |
Affymetrix Human Clariom D Assay | PSR0300148412.hg.1 | ENST00000425459.5 | Ensembl |
Affymetrix Human Clariom D Assay | PSR0300148412.hg.1 | ENST00000428684.1 | Ensembl |
Affymetrix Human Clariom D Assay | PSR0300148412.hg.1 | ENST00000429432.5 | Ensembl |
Affymetrix Human Clariom D Assay | PSR0300148412.hg.1 | ENST00000431009.1 | Ensembl |
Affymetrix Human Clariom D Assay | PSR0300148412.hg.1 | NM_015141.3 | Refseq |
Affymetrix Human Clariom D Assay | PSR0300148412.hg.1 | ENST00000282541.9 | Ensembl |
Affymetrix Human Clariom D Assay | PSR0300148412.hg.1 | ENST00000425459.5 | Ensembl |
Affymetrix Human Clariom D Assay | PSR0300148412.hg.1 | ENST00000428684.1 | Ensembl |
Affymetrix Human Clariom D Assay | PSR0300148412.hg.1 | ENST00000429432.5 | Ensembl |
Affymetrix Human Clariom D Assay | PSR0300148412.hg.1 | ENST00000431009.1 | Ensembl |
Affymetrix Human Clariom D Assay | PSR0300148412.hg.1 | NM_015141.3 | Refseq |
Affymetrix Human Clariom D Assay | PSR0300148412.hg.1 | TC0300006956.hg.1 | Clariom D TC |
