Assay Details
| Gene Symbol: RNF114 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | NM_018683.3 | NP_061153.1 | 3 - 4 | 435 | 62 | |
| GenBank mRNA | AK091976.1 | - | 3 - 4 | 413 | 62 | |
| AK097265.1 | - | 1 - 2 | 1559 | 62 | ||
| AK301731.1 | - | 3 - 4 | 405 | 62 | ||
| AK315638.1 | - | 3 - 4 | 415 | 62 | ||
| BC013695.1 | - | 3 - 4 | 404 | 3858611 | 62 | |
| BC066919.1 | - | 3 - 4 | 413 | 5264729 | 62 | |
| BM830132.1 | - | 3 - 4 | 433 | 62 | ||
| BT006795.1 | - | 3 - 4 | 399 | 62 | ||
| BX640603.1 | - | 3 - 4 | 417 | 62 | ||
More Information
Updated Mapping Information
This assay targets the same location as HTA transcript cluster(s) TC20000393.hg.1
Set Membership: |
|
Matching Ambion siRNAs:
Gene Ontology Categories:
Function(s)
Process(es)
protein polyubiquitination
multicellular organism development
spermatogenesis
cell differentiation
positive regulation of proteasomal ubiquitin-dependent protein catabolic process
ubiquitin-protein transferase activity
zinc ion binding
ligase activity
ubiquitin conjugating enzyme binding
ubiquitin protein ligase activity
protein polyubiquitination
multicellular organism development
spermatogenesis
cell differentiation
positive regulation of proteasomal ubiquitin-dependent protein catabolic process
ubiquitin-protein transferase activity
zinc ion binding
ligase activity
ubiquitin conjugating enzyme binding
ubiquitin protein ligase activity
MicroArray Probes:
Affymetrix
| Array Name | Probe IDs | Targets | Target Type |
|---|---|---|---|
| Affymetrix Human Transcriptome Array 2.0 | TC2000007666.hg.1 | ENST00000244061.5 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC2000007666.hg.1 | ENST00000244061.5 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC2000007666.hg.1 | ENST00000244061.5 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC2000007666.hg.1 | ENST00000244061.5 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC2000007666.hg.1 | ENST00000244061.5 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC2000007666.hg.1 | ENST00000244061.5 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC2000007666.hg.1 | ENST00000244061.5 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC2000007666.hg.1 | ENST00000244061.5 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC2000007666.hg.1 | ENST00000244061.5 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC2000007666.hg.1 | ENST00000244061.5 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC2000007666.hg.1 | ENST00000244061.5 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC2000007666.hg.1 | ENST00000622920.1 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC2000007666.hg.1 | ENST00000622920.1 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC2000007666.hg.1 | ENST00000622920.1 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC2000007666.hg.1 | ENST00000622920.1 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC2000007666.hg.1 | ENST00000622920.1 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC2000007666.hg.1 | ENST00000622920.1 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC2000007666.hg.1 | ENST00000622920.1 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC2000007666.hg.1 | ENST00000622999.3 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC2000007666.hg.1 | ENST00000622999.3 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC2000007666.hg.1 | ENST00000622999.3 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC2000007666.hg.1 | ENST00000622999.3 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC2000007666.hg.1 | ENST00000622999.3 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC2000007666.hg.1 | ENST00000622999.3 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC2000007666.hg.1 | ENST00000622999.3 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC2000007666.hg.1 | ENST00000622999.3 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC2000007666.hg.1 | ENST00000622999.3 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC2000007666.hg.1 | ENST00000622999.3 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC2000007666.hg.1 | ENST00000623528.3 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC2000007666.hg.1 | ENST00000623528.3 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC2000007666.hg.1 | ENST00000623528.3 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC2000007666.hg.1 | ENST00000623528.3 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC2000007666.hg.1 | ENST00000623528.3 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC2000007666.hg.1 | ENST00000623528.3 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC2000007666.hg.1 | ENST00000623528.3 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC2000007666.hg.1 | ENST00000623732.3 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC2000007666.hg.1 | ENST00000623732.3 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC2000007666.hg.1 | ENST00000623732.3 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC2000007666.hg.1 | ENST00000623732.3 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC2000007666.hg.1 | ENST00000623732.3 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC2000007666.hg.1 | ENST00000624620.1 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC2000007666.hg.1 | ENST00000624620.1 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC2000007666.hg.1 | ENST00000624620.1 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC2000007666.hg.1 | ENST00000624666.1 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC2000007666.hg.1 | ENST00000624666.1 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC2000007666.hg.1 | ENST00000624666.1 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC2000007666.hg.1 | ENST00000624666.1 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC2000007666.hg.1 | ENST00000624666.1 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC2000007666.hg.1 | ENST00000624666.1 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC2000007666.hg.1 | ENST00000624666.1 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC2000007666.hg.1 | ENST00000625177.3 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC2000007666.hg.1 | ENST00000625177.3 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC2000007666.hg.1 | ENST00000625177.3 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC2000007666.hg.1 | ENST00000625177.3 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC2000007666.hg.1 | ENST00000625177.3 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC2000007666.hg.1 | ENST00000625177.3 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC2000007666.hg.1 | NM_018683.3 | Refseq |
| Affymetrix Human Transcriptome Array 2.0 | TC2000007666.hg.1 | NM_018683.3 | Refseq |
| Affymetrix Human Transcriptome Array 2.0 | TC2000007666.hg.1 | NM_018683.3 | Refseq |
| Affymetrix Human Transcriptome Array 2.0 | TC2000007666.hg.1 | NM_018683.3 | Refseq |
| Affymetrix Human Transcriptome Array 2.0 | TC2000007666.hg.1 | NM_018683.3 | Refseq |
| Affymetrix Human Transcriptome Array 2.0 | TC2000007666.hg.1 | NM_018683.3 | Refseq |
| Affymetrix Human Transcriptome Array 2.0 | TC2000007666.hg.1 | NM_018683.3 | Refseq |
| Affymetrix Human Transcriptome Array 2.0 | TC2000007666.hg.1 | NM_018683.3 | Refseq |
| Affymetrix Human Transcriptome Array 2.0 | TC2000007666.hg.1 | NM_018683.3 | Refseq |
| Affymetrix Human Transcriptome Array 2.0 | TC2000007666.hg.1 | NM_018683.3 | Refseq |
| Affymetrix Human Transcriptome Array 2.0 | PSR20005675.hg.1 | ENST00000244061.2 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | PSR20005675.hg.1 | ENST00000449816.2 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | PSR20005675.hg.1 | NM_018683.3 | Refseq |
| Affymetrix Human Transcriptome Array 2.0 | PSR20005675.hg.1 | TC20000393.hg.1 | HTA TC |
| Affymetrix Human Clariom D Assay | PSR2000153471.hg.1 | ENST00000244061.5 | Ensembl |
| Affymetrix Human Clariom D Assay | PSR2000153471.hg.1 | ENST00000622920.1 | Ensembl |
| Affymetrix Human Clariom D Assay | PSR2000153471.hg.1 | ENST00000622999.3 | Ensembl |
| Affymetrix Human Clariom D Assay | PSR2000153471.hg.1 | ENST00000623528.3 | Ensembl |
| Affymetrix Human Clariom D Assay | PSR2000153471.hg.1 | ENST00000623732.3 | Ensembl |
| Affymetrix Human Clariom D Assay | PSR2000153471.hg.1 | ENST00000624666.1 | Ensembl |
| Affymetrix Human Clariom D Assay | PSR2000153471.hg.1 | ENST00000625177.3 | Ensembl |
| Affymetrix Human Clariom D Assay | PSR2000153471.hg.1 | NM_018683.3 | Refseq |
| Affymetrix Human Clariom D Assay | PSR2000153471.hg.1 | TC2000007666.hg.1 | Clariom D TC |
| Affymetrix Human Clariom D Assay | PSR2000153471.hg.1 | ENST00000244061.5 | Ensembl |
| Affymetrix Human Clariom D Assay | PSR2000153471.hg.1 | ENST00000622920.1 | Ensembl |
| Affymetrix Human Clariom D Assay | PSR2000153471.hg.1 | ENST00000622999.3 | Ensembl |
| Affymetrix Human Clariom D Assay | PSR2000153471.hg.1 | ENST00000623528.3 | Ensembl |
| Affymetrix Human Clariom D Assay | PSR2000153471.hg.1 | ENST00000623732.3 | Ensembl |
| Affymetrix Human Clariom D Assay | PSR2000153471.hg.1 | ENST00000624666.1 | Ensembl |
| Affymetrix Human Clariom D Assay | PSR2000153471.hg.1 | ENST00000625177.3 | Ensembl |
| Affymetrix Human Clariom D Assay | PSR2000153471.hg.1 | NM_018683.3 | Refseq |
| Affymetrix Human Clariom D Assay | TC2000007666.hg.1 | ENST00000244061.5 | Ensembl |
| Affymetrix Human Clariom D Assay | TC2000007666.hg.1 | ENST00000244061.5 | Ensembl |
| Affymetrix Human Clariom D Assay | TC2000007666.hg.1 | ENST00000244061.5 | Ensembl |
| Affymetrix Human Clariom D Assay | TC2000007666.hg.1 | ENST00000244061.5 | Ensembl |
| Affymetrix Human Clariom D Assay | TC2000007666.hg.1 | ENST00000244061.5 | Ensembl |
| Affymetrix Human Clariom D Assay | TC2000007666.hg.1 | ENST00000244061.5 | Ensembl |
| Affymetrix Human Clariom D Assay | TC2000007666.hg.1 | ENST00000244061.5 | Ensembl |
| Affymetrix Human Clariom D Assay | TC2000007666.hg.1 | ENST00000244061.5 | Ensembl |
| Affymetrix Human Clariom D Assay | TC2000007666.hg.1 | ENST00000244061.5 | Ensembl |
| Affymetrix Human Clariom D Assay | TC2000007666.hg.1 | ENST00000244061.5 | Ensembl |
| Affymetrix Human Clariom D Assay | TC2000007666.hg.1 | ENST00000244061.5 | Ensembl |
| Affymetrix Human Clariom D Assay | TC2000007666.hg.1 | ENST00000622920.1 | Ensembl |
| Affymetrix Human Clariom D Assay | TC2000007666.hg.1 | ENST00000622920.1 | Ensembl |
| Affymetrix Human Clariom D Assay | TC2000007666.hg.1 | ENST00000622920.1 | Ensembl |
| Affymetrix Human Clariom D Assay | TC2000007666.hg.1 | ENST00000622920.1 | Ensembl |
| Affymetrix Human Clariom D Assay | TC2000007666.hg.1 | ENST00000622920.1 | Ensembl |
| Affymetrix Human Clariom D Assay | TC2000007666.hg.1 | ENST00000622920.1 | Ensembl |
| Affymetrix Human Clariom D Assay | TC2000007666.hg.1 | ENST00000622920.1 | Ensembl |
| Affymetrix Human Clariom D Assay | TC2000007666.hg.1 | ENST00000622999.3 | Ensembl |
| Affymetrix Human Clariom D Assay | TC2000007666.hg.1 | ENST00000622999.3 | Ensembl |
| Affymetrix Human Clariom D Assay | TC2000007666.hg.1 | ENST00000622999.3 | Ensembl |
| Affymetrix Human Clariom D Assay | TC2000007666.hg.1 | ENST00000622999.3 | Ensembl |
| Affymetrix Human Clariom D Assay | TC2000007666.hg.1 | ENST00000622999.3 | Ensembl |
| Affymetrix Human Clariom D Assay | TC2000007666.hg.1 | ENST00000622999.3 | Ensembl |
| Affymetrix Human Clariom D Assay | TC2000007666.hg.1 | ENST00000622999.3 | Ensembl |
| Affymetrix Human Clariom D Assay | TC2000007666.hg.1 | ENST00000622999.3 | Ensembl |
| Affymetrix Human Clariom D Assay | TC2000007666.hg.1 | ENST00000622999.3 | Ensembl |
| Affymetrix Human Clariom D Assay | TC2000007666.hg.1 | ENST00000622999.3 | Ensembl |
| Affymetrix Human Clariom D Assay | TC2000007666.hg.1 | ENST00000623528.3 | Ensembl |
| Affymetrix Human Clariom D Assay | TC2000007666.hg.1 | ENST00000623528.3 | Ensembl |
| Affymetrix Human Clariom D Assay | TC2000007666.hg.1 | ENST00000623528.3 | Ensembl |
| Affymetrix Human Clariom D Assay | TC2000007666.hg.1 | ENST00000623528.3 | Ensembl |
| Affymetrix Human Clariom D Assay | TC2000007666.hg.1 | ENST00000623528.3 | Ensembl |
| Affymetrix Human Clariom D Assay | TC2000007666.hg.1 | ENST00000623528.3 | Ensembl |
| Affymetrix Human Clariom D Assay | TC2000007666.hg.1 | ENST00000623528.3 | Ensembl |
| Affymetrix Human Clariom D Assay | TC2000007666.hg.1 | ENST00000623732.3 | Ensembl |
| Affymetrix Human Clariom D Assay | TC2000007666.hg.1 | ENST00000623732.3 | Ensembl |
| Affymetrix Human Clariom D Assay | TC2000007666.hg.1 | ENST00000623732.3 | Ensembl |
| Affymetrix Human Clariom D Assay | TC2000007666.hg.1 | ENST00000623732.3 | Ensembl |
| Affymetrix Human Clariom D Assay | TC2000007666.hg.1 | ENST00000623732.3 | Ensembl |
| Affymetrix Human Clariom D Assay | TC2000007666.hg.1 | ENST00000624620.1 | Ensembl |
| Affymetrix Human Clariom D Assay | TC2000007666.hg.1 | ENST00000624620.1 | Ensembl |
| Affymetrix Human Clariom D Assay | TC2000007666.hg.1 | ENST00000624620.1 | Ensembl |
| Affymetrix Human Clariom D Assay | TC2000007666.hg.1 | ENST00000624666.1 | Ensembl |
| Affymetrix Human Clariom D Assay | TC2000007666.hg.1 | ENST00000624666.1 | Ensembl |
| Affymetrix Human Clariom D Assay | TC2000007666.hg.1 | ENST00000624666.1 | Ensembl |
| Affymetrix Human Clariom D Assay | TC2000007666.hg.1 | ENST00000624666.1 | Ensembl |
| Affymetrix Human Clariom D Assay | TC2000007666.hg.1 | ENST00000624666.1 | Ensembl |
| Affymetrix Human Clariom D Assay | TC2000007666.hg.1 | ENST00000624666.1 | Ensembl |
| Affymetrix Human Clariom D Assay | TC2000007666.hg.1 | ENST00000624666.1 | Ensembl |
| Affymetrix Human Clariom D Assay | TC2000007666.hg.1 | ENST00000625177.3 | Ensembl |
| Affymetrix Human Clariom D Assay | TC2000007666.hg.1 | ENST00000625177.3 | Ensembl |
| Affymetrix Human Clariom D Assay | TC2000007666.hg.1 | ENST00000625177.3 | Ensembl |
| Affymetrix Human Clariom D Assay | TC2000007666.hg.1 | ENST00000625177.3 | Ensembl |
| Affymetrix Human Clariom D Assay | TC2000007666.hg.1 | ENST00000625177.3 | Ensembl |
| Affymetrix Human Clariom D Assay | TC2000007666.hg.1 | ENST00000625177.3 | Ensembl |
| Affymetrix Human Clariom D Assay | TC2000007666.hg.1 | NM_018683.3 | Refseq |
| Affymetrix Human Clariom D Assay | TC2000007666.hg.1 | NM_018683.3 | Refseq |
| Affymetrix Human Clariom D Assay | TC2000007666.hg.1 | NM_018683.3 | Refseq |
| Affymetrix Human Clariom D Assay | TC2000007666.hg.1 | NM_018683.3 | Refseq |
| Affymetrix Human Clariom D Assay | TC2000007666.hg.1 | NM_018683.3 | Refseq |
| Affymetrix Human Clariom D Assay | TC2000007666.hg.1 | NM_018683.3 | Refseq |
| Affymetrix Human Clariom D Assay | TC2000007666.hg.1 | NM_018683.3 | Refseq |
| Affymetrix Human Clariom D Assay | TC2000007666.hg.1 | NM_018683.3 | Refseq |
| Affymetrix Human Clariom D Assay | TC2000007666.hg.1 | NM_018683.3 | Refseq |
| Affymetrix Human Clariom D Assay | TC2000007666.hg.1 | NM_018683.3 | Refseq |
| Affymetrix Human Clariom D Assay | PSR2000153472.hg.1 | ENST00000244061.5 | Ensembl |
| Affymetrix Human Clariom D Assay | PSR2000153472.hg.1 | ENST00000622920.1 | Ensembl |
| Affymetrix Human Clariom D Assay | PSR2000153472.hg.1 | ENST00000622999.3 | Ensembl |
| Affymetrix Human Clariom D Assay | PSR2000153472.hg.1 | ENST00000623528.3 | Ensembl |
| Affymetrix Human Clariom D Assay | PSR2000153472.hg.1 | ENST00000624666.1 | Ensembl |
| Affymetrix Human Clariom D Assay | PSR2000153472.hg.1 | ENST00000625177.3 | Ensembl |
| Affymetrix Human Clariom D Assay | PSR2000153472.hg.1 | NM_018683.3 | Refseq |
| Affymetrix Human Clariom D Assay | PSR2000153472.hg.1 | TC2000007666.hg.1 | Clariom D TC |
| Affymetrix Human Clariom D Assay | PSR2000153472.hg.1 | ENST00000244061.5 | Ensembl |
| Affymetrix Human Clariom D Assay | PSR2000153472.hg.1 | ENST00000622920.1 | Ensembl |
| Affymetrix Human Clariom D Assay | PSR2000153472.hg.1 | ENST00000622999.3 | Ensembl |
| Affymetrix Human Clariom D Assay | PSR2000153472.hg.1 | ENST00000623528.3 | Ensembl |
| Affymetrix Human Clariom D Assay | PSR2000153472.hg.1 | ENST00000624666.1 | Ensembl |
| Affymetrix Human Clariom D Assay | PSR2000153472.hg.1 | ENST00000625177.3 | Ensembl |
| Affymetrix Human Clariom D Assay | PSR2000153472.hg.1 | NM_018683.3 | Refseq |

RNF114
Genomic Map