Assay Details
| Gene Symbol: LOC100507291 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | NR_121608.1 | 3 - 4 | 355 | 96 | ||
| GenBank mRNA | BX098847.1 | - | 3 - 4 | 349 | 96 | |
| DB034152.1 | - | 3 - 4 | 355 | 96 | ||
| DB446595.1 | - | 3 - 4 | 356 | 96 | ||
More Information
Set Membership: |
|

LOC100507291
Genomic Map