Assay Details
| Gene Symbol: BIRC5 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| GenBank mRNA | AB154416.1 | - | 3 - 4 | 343 | 78 | |
| DQ508245.1 | - | 4 - 5 | 348 | 78 | ||
| DQ508246.1 | - | 4 - 5 | 344 | 78 | ||
| DQ508248.1 | - | 4 - 5 | 376 | 78 | ||
| DQ508249.1 | - | 3 - 4 | 993 | 78 | ||
More Information
Set Membership: |
|
MicroArray Probes:
Affymetrix
| Array Name | Probe IDs | Targets | Target Type |
|---|---|---|---|
| Affymetrix Human Transcriptome Array 2.0 | TC1700009043.hg.1 | ENST00000301633.8 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC1700009043.hg.1 | ENST00000301633.8 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC1700009043.hg.1 | ENST00000301633.8 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC1700009043.hg.1 | ENST00000301633.8 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC1700009043.hg.1 | ENST00000301633.8 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC1700009043.hg.1 | ENST00000301633.8 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC1700009043.hg.1 | ENST00000301633.8 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC1700009043.hg.1 | ENST00000301633.8 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC1700009043.hg.1 | ENST00000301633.8 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC1700009043.hg.1 | ENST00000301633.8 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC1700009043.hg.1 | ENST00000301633.8 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC1700009043.hg.1 | ENST00000350051.7 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC1700009043.hg.1 | ENST00000350051.7 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC1700009043.hg.1 | ENST00000350051.7 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC1700009043.hg.1 | ENST00000350051.7 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC1700009043.hg.1 | ENST00000350051.7 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC1700009043.hg.1 | ENST00000350051.7 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC1700009043.hg.1 | ENST00000350051.7 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC1700009043.hg.1 | ENST00000350051.7 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC1700009043.hg.1 | ENST00000350051.7 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC1700009043.hg.1 | ENST00000350051.7 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC1700009043.hg.1 | ENST00000374948.6 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC1700009043.hg.1 | ENST00000374948.6 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC1700009043.hg.1 | ENST00000374948.6 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC1700009043.hg.1 | ENST00000374948.6 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC1700009043.hg.1 | ENST00000374948.6 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC1700009043.hg.1 | ENST00000374948.6 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC1700009043.hg.1 | ENST00000374948.6 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC1700009043.hg.1 | ENST00000374948.6 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC1700009043.hg.1 | ENST00000586192.5 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC1700009043.hg.1 | ENST00000586192.5 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC1700009043.hg.1 | ENST00000586192.5 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC1700009043.hg.1 | ENST00000586192.5 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC1700009043.hg.1 | ENST00000586192.5 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC1700009043.hg.1 | ENST00000586192.5 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC1700009043.hg.1 | ENST00000587746.5 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC1700009043.hg.1 | ENST00000587746.5 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC1700009043.hg.1 | ENST00000587746.5 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC1700009043.hg.1 | ENST00000587746.5 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC1700009043.hg.1 | ENST00000587746.5 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC1700009043.hg.1 | ENST00000589892.1 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC1700009043.hg.1 | ENST00000589892.1 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC1700009043.hg.1 | ENST00000589892.1 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC1700009043.hg.1 | ENST00000589892.1 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC1700009043.hg.1 | ENST00000589892.1 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC1700009043.hg.1 | ENST00000590449.1 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC1700009043.hg.1 | ENST00000590449.1 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC1700009043.hg.1 | ENST00000590449.1 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC1700009043.hg.1 | ENST00000590449.1 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC1700009043.hg.1 | ENST00000590925.6 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC1700009043.hg.1 | ENST00000590925.6 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC1700009043.hg.1 | ENST00000590925.6 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC1700009043.hg.1 | ENST00000590925.6 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC1700009043.hg.1 | ENST00000590925.6 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC1700009043.hg.1 | ENST00000590925.6 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC1700009043.hg.1 | ENST00000591800.1 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC1700009043.hg.1 | ENST00000591800.1 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC1700009043.hg.1 | ENST00000591800.1 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC1700009043.hg.1 | ENST00000591800.1 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC1700009043.hg.1 | ENST00000591800.1 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC1700009043.hg.1 | ENST00000592115.5 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC1700009043.hg.1 | ENST00000592115.5 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC1700009043.hg.1 | ENST00000592115.5 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC1700009043.hg.1 | ENST00000592115.5 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC1700009043.hg.1 | ENST00000592115.5 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC1700009043.hg.1 | ENST00000592115.5 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC1700009043.hg.1 | ENST00000592115.5 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC1700009043.hg.1 | ENST00000592115.5 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC1700009043.hg.1 | ENST00000592734.5 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC1700009043.hg.1 | ENST00000592734.5 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC1700009043.hg.1 | ENST00000592734.5 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC1700009043.hg.1 | ENST00000592734.5 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC1700009043.hg.1 | NM_001012270.1 | Refseq |
| Affymetrix Human Transcriptome Array 2.0 | TC1700009043.hg.1 | NM_001012270.1 | Refseq |
| Affymetrix Human Transcriptome Array 2.0 | TC1700009043.hg.1 | NM_001012270.1 | Refseq |
| Affymetrix Human Transcriptome Array 2.0 | TC1700009043.hg.1 | NM_001012270.1 | Refseq |
| Affymetrix Human Transcriptome Array 2.0 | TC1700009043.hg.1 | NM_001012270.1 | Refseq |
| Affymetrix Human Transcriptome Array 2.0 | TC1700009043.hg.1 | NM_001012270.1 | Refseq |
| Affymetrix Human Transcriptome Array 2.0 | TC1700009043.hg.1 | NM_001012270.1 | Refseq |
| Affymetrix Human Transcriptome Array 2.0 | TC1700009043.hg.1 | NM_001012270.1 | Refseq |
| Affymetrix Human Transcriptome Array 2.0 | TC1700009043.hg.1 | NM_001012270.1 | Refseq |
| Affymetrix Human Transcriptome Array 2.0 | TC1700009043.hg.1 | NM_001012271.1 | Refseq |
| Affymetrix Human Transcriptome Array 2.0 | TC1700009043.hg.1 | NM_001012271.1 | Refseq |
| Affymetrix Human Transcriptome Array 2.0 | TC1700009043.hg.1 | NM_001012271.1 | Refseq |
| Affymetrix Human Transcriptome Array 2.0 | TC1700009043.hg.1 | NM_001012271.1 | Refseq |
| Affymetrix Human Transcriptome Array 2.0 | TC1700009043.hg.1 | NM_001012271.1 | Refseq |
| Affymetrix Human Transcriptome Array 2.0 | TC1700009043.hg.1 | NM_001012271.1 | Refseq |
| Affymetrix Human Transcriptome Array 2.0 | TC1700009043.hg.1 | NM_001012271.1 | Refseq |
| Affymetrix Human Transcriptome Array 2.0 | TC1700009043.hg.1 | NM_001012271.1 | Refseq |
| Affymetrix Human Transcriptome Array 2.0 | TC1700009043.hg.1 | NM_001012271.1 | Refseq |
| Affymetrix Human Transcriptome Array 2.0 | TC1700009043.hg.1 | NM_001012271.1 | Refseq |
| Affymetrix Human Transcriptome Array 2.0 | TC1700009043.hg.1 | NM_001012271.1 | Refseq |
| Affymetrix Human Transcriptome Array 2.0 | TC1700009043.hg.1 | NM_001168.2 | Refseq |
| Affymetrix Human Transcriptome Array 2.0 | TC1700009043.hg.1 | NM_001168.2 | Refseq |
| Affymetrix Human Transcriptome Array 2.0 | TC1700009043.hg.1 | NM_001168.2 | Refseq |
| Affymetrix Human Transcriptome Array 2.0 | TC1700009043.hg.1 | NM_001168.2 | Refseq |
| Affymetrix Human Transcriptome Array 2.0 | TC1700009043.hg.1 | NM_001168.2 | Refseq |
| Affymetrix Human Transcriptome Array 2.0 | TC1700009043.hg.1 | NM_001168.2 | Refseq |
| Affymetrix Human Transcriptome Array 2.0 | TC1700009043.hg.1 | NM_001168.2 | Refseq |
| Affymetrix Human Transcriptome Array 2.0 | TC1700009043.hg.1 | NM_001168.2 | Refseq |
| Affymetrix Human Transcriptome Array 2.0 | TC1700009043.hg.1 | NM_001168.2 | Refseq |
| Affymetrix Human Transcriptome Array 2.0 | TC1700009043.hg.1 | NM_001168.2 | Refseq |
| Affymetrix Human Clariom D Assay | TC1700009043.hg.1 | ENST00000301633.8 | Ensembl |
| Affymetrix Human Clariom D Assay | TC1700009043.hg.1 | ENST00000301633.8 | Ensembl |
| Affymetrix Human Clariom D Assay | TC1700009043.hg.1 | ENST00000301633.8 | Ensembl |
| Affymetrix Human Clariom D Assay | TC1700009043.hg.1 | ENST00000301633.8 | Ensembl |
| Affymetrix Human Clariom D Assay | TC1700009043.hg.1 | ENST00000301633.8 | Ensembl |
| Affymetrix Human Clariom D Assay | TC1700009043.hg.1 | ENST00000301633.8 | Ensembl |
| Affymetrix Human Clariom D Assay | TC1700009043.hg.1 | ENST00000301633.8 | Ensembl |
| Affymetrix Human Clariom D Assay | TC1700009043.hg.1 | ENST00000301633.8 | Ensembl |
| Affymetrix Human Clariom D Assay | TC1700009043.hg.1 | ENST00000301633.8 | Ensembl |
| Affymetrix Human Clariom D Assay | TC1700009043.hg.1 | ENST00000301633.8 | Ensembl |
| Affymetrix Human Clariom D Assay | TC1700009043.hg.1 | ENST00000301633.8 | Ensembl |
| Affymetrix Human Clariom D Assay | TC1700009043.hg.1 | ENST00000350051.7 | Ensembl |
| Affymetrix Human Clariom D Assay | TC1700009043.hg.1 | ENST00000350051.7 | Ensembl |
| Affymetrix Human Clariom D Assay | TC1700009043.hg.1 | ENST00000350051.7 | Ensembl |
| Affymetrix Human Clariom D Assay | TC1700009043.hg.1 | ENST00000350051.7 | Ensembl |
| Affymetrix Human Clariom D Assay | TC1700009043.hg.1 | ENST00000350051.7 | Ensembl |
| Affymetrix Human Clariom D Assay | TC1700009043.hg.1 | ENST00000350051.7 | Ensembl |
| Affymetrix Human Clariom D Assay | TC1700009043.hg.1 | ENST00000350051.7 | Ensembl |
| Affymetrix Human Clariom D Assay | TC1700009043.hg.1 | ENST00000350051.7 | Ensembl |
| Affymetrix Human Clariom D Assay | TC1700009043.hg.1 | ENST00000350051.7 | Ensembl |
| Affymetrix Human Clariom D Assay | TC1700009043.hg.1 | ENST00000350051.7 | Ensembl |
| Affymetrix Human Clariom D Assay | TC1700009043.hg.1 | ENST00000374948.6 | Ensembl |
| Affymetrix Human Clariom D Assay | TC1700009043.hg.1 | ENST00000374948.6 | Ensembl |
| Affymetrix Human Clariom D Assay | TC1700009043.hg.1 | ENST00000374948.6 | Ensembl |
| Affymetrix Human Clariom D Assay | TC1700009043.hg.1 | ENST00000374948.6 | Ensembl |
| Affymetrix Human Clariom D Assay | TC1700009043.hg.1 | ENST00000374948.6 | Ensembl |
| Affymetrix Human Clariom D Assay | TC1700009043.hg.1 | ENST00000374948.6 | Ensembl |
| Affymetrix Human Clariom D Assay | TC1700009043.hg.1 | ENST00000374948.6 | Ensembl |
| Affymetrix Human Clariom D Assay | TC1700009043.hg.1 | ENST00000374948.6 | Ensembl |
| Affymetrix Human Clariom D Assay | TC1700009043.hg.1 | ENST00000586192.5 | Ensembl |
| Affymetrix Human Clariom D Assay | TC1700009043.hg.1 | ENST00000586192.5 | Ensembl |
| Affymetrix Human Clariom D Assay | TC1700009043.hg.1 | ENST00000586192.5 | Ensembl |
| Affymetrix Human Clariom D Assay | TC1700009043.hg.1 | ENST00000586192.5 | Ensembl |
| Affymetrix Human Clariom D Assay | TC1700009043.hg.1 | ENST00000586192.5 | Ensembl |
| Affymetrix Human Clariom D Assay | TC1700009043.hg.1 | ENST00000586192.5 | Ensembl |
| Affymetrix Human Clariom D Assay | TC1700009043.hg.1 | ENST00000587746.5 | Ensembl |
| Affymetrix Human Clariom D Assay | TC1700009043.hg.1 | ENST00000587746.5 | Ensembl |
| Affymetrix Human Clariom D Assay | TC1700009043.hg.1 | ENST00000587746.5 | Ensembl |
| Affymetrix Human Clariom D Assay | TC1700009043.hg.1 | ENST00000587746.5 | Ensembl |
| Affymetrix Human Clariom D Assay | TC1700009043.hg.1 | ENST00000587746.5 | Ensembl |
| Affymetrix Human Clariom D Assay | TC1700009043.hg.1 | ENST00000589892.1 | Ensembl |
| Affymetrix Human Clariom D Assay | TC1700009043.hg.1 | ENST00000589892.1 | Ensembl |
| Affymetrix Human Clariom D Assay | TC1700009043.hg.1 | ENST00000589892.1 | Ensembl |
| Affymetrix Human Clariom D Assay | TC1700009043.hg.1 | ENST00000589892.1 | Ensembl |
| Affymetrix Human Clariom D Assay | TC1700009043.hg.1 | ENST00000589892.1 | Ensembl |
| Affymetrix Human Clariom D Assay | TC1700009043.hg.1 | ENST00000590449.1 | Ensembl |
| Affymetrix Human Clariom D Assay | TC1700009043.hg.1 | ENST00000590449.1 | Ensembl |
| Affymetrix Human Clariom D Assay | TC1700009043.hg.1 | ENST00000590449.1 | Ensembl |
| Affymetrix Human Clariom D Assay | TC1700009043.hg.1 | ENST00000590449.1 | Ensembl |
| Affymetrix Human Clariom D Assay | TC1700009043.hg.1 | ENST00000590925.6 | Ensembl |
| Affymetrix Human Clariom D Assay | TC1700009043.hg.1 | ENST00000590925.6 | Ensembl |
| Affymetrix Human Clariom D Assay | TC1700009043.hg.1 | ENST00000590925.6 | Ensembl |
| Affymetrix Human Clariom D Assay | TC1700009043.hg.1 | ENST00000590925.6 | Ensembl |
| Affymetrix Human Clariom D Assay | TC1700009043.hg.1 | ENST00000590925.6 | Ensembl |
| Affymetrix Human Clariom D Assay | TC1700009043.hg.1 | ENST00000590925.6 | Ensembl |
| Affymetrix Human Clariom D Assay | TC1700009043.hg.1 | ENST00000591800.1 | Ensembl |
| Affymetrix Human Clariom D Assay | TC1700009043.hg.1 | ENST00000591800.1 | Ensembl |
| Affymetrix Human Clariom D Assay | TC1700009043.hg.1 | ENST00000591800.1 | Ensembl |
| Affymetrix Human Clariom D Assay | TC1700009043.hg.1 | ENST00000591800.1 | Ensembl |
| Affymetrix Human Clariom D Assay | TC1700009043.hg.1 | ENST00000591800.1 | Ensembl |
| Affymetrix Human Clariom D Assay | TC1700009043.hg.1 | ENST00000592115.5 | Ensembl |
| Affymetrix Human Clariom D Assay | TC1700009043.hg.1 | ENST00000592115.5 | Ensembl |
| Affymetrix Human Clariom D Assay | TC1700009043.hg.1 | ENST00000592115.5 | Ensembl |
| Affymetrix Human Clariom D Assay | TC1700009043.hg.1 | ENST00000592115.5 | Ensembl |
| Affymetrix Human Clariom D Assay | TC1700009043.hg.1 | ENST00000592115.5 | Ensembl |
| Affymetrix Human Clariom D Assay | TC1700009043.hg.1 | ENST00000592115.5 | Ensembl |
| Affymetrix Human Clariom D Assay | TC1700009043.hg.1 | ENST00000592115.5 | Ensembl |
| Affymetrix Human Clariom D Assay | TC1700009043.hg.1 | ENST00000592115.5 | Ensembl |
| Affymetrix Human Clariom D Assay | TC1700009043.hg.1 | ENST00000592734.5 | Ensembl |
| Affymetrix Human Clariom D Assay | TC1700009043.hg.1 | ENST00000592734.5 | Ensembl |
| Affymetrix Human Clariom D Assay | TC1700009043.hg.1 | ENST00000592734.5 | Ensembl |
| Affymetrix Human Clariom D Assay | TC1700009043.hg.1 | ENST00000592734.5 | Ensembl |
| Affymetrix Human Clariom D Assay | TC1700009043.hg.1 | NM_001012270.1 | Refseq |
| Affymetrix Human Clariom D Assay | TC1700009043.hg.1 | NM_001012270.1 | Refseq |
| Affymetrix Human Clariom D Assay | TC1700009043.hg.1 | NM_001012270.1 | Refseq |
| Affymetrix Human Clariom D Assay | TC1700009043.hg.1 | NM_001012270.1 | Refseq |
| Affymetrix Human Clariom D Assay | TC1700009043.hg.1 | NM_001012270.1 | Refseq |
| Affymetrix Human Clariom D Assay | TC1700009043.hg.1 | NM_001012270.1 | Refseq |
| Affymetrix Human Clariom D Assay | TC1700009043.hg.1 | NM_001012270.1 | Refseq |
| Affymetrix Human Clariom D Assay | TC1700009043.hg.1 | NM_001012270.1 | Refseq |
| Affymetrix Human Clariom D Assay | TC1700009043.hg.1 | NM_001012270.1 | Refseq |
| Affymetrix Human Clariom D Assay | TC1700009043.hg.1 | NM_001012271.1 | Refseq |
| Affymetrix Human Clariom D Assay | TC1700009043.hg.1 | NM_001012271.1 | Refseq |
| Affymetrix Human Clariom D Assay | TC1700009043.hg.1 | NM_001012271.1 | Refseq |
| Affymetrix Human Clariom D Assay | TC1700009043.hg.1 | NM_001012271.1 | Refseq |
| Affymetrix Human Clariom D Assay | TC1700009043.hg.1 | NM_001012271.1 | Refseq |
| Affymetrix Human Clariom D Assay | TC1700009043.hg.1 | NM_001012271.1 | Refseq |
| Affymetrix Human Clariom D Assay | TC1700009043.hg.1 | NM_001012271.1 | Refseq |
| Affymetrix Human Clariom D Assay | TC1700009043.hg.1 | NM_001012271.1 | Refseq |
| Affymetrix Human Clariom D Assay | TC1700009043.hg.1 | NM_001012271.1 | Refseq |
| Affymetrix Human Clariom D Assay | TC1700009043.hg.1 | NM_001012271.1 | Refseq |
| Affymetrix Human Clariom D Assay | TC1700009043.hg.1 | NM_001012271.1 | Refseq |
| Affymetrix Human Clariom D Assay | TC1700009043.hg.1 | NM_001168.2 | Refseq |
| Affymetrix Human Clariom D Assay | TC1700009043.hg.1 | NM_001168.2 | Refseq |
| Affymetrix Human Clariom D Assay | TC1700009043.hg.1 | NM_001168.2 | Refseq |
| Affymetrix Human Clariom D Assay | TC1700009043.hg.1 | NM_001168.2 | Refseq |
| Affymetrix Human Clariom D Assay | TC1700009043.hg.1 | NM_001168.2 | Refseq |
| Affymetrix Human Clariom D Assay | TC1700009043.hg.1 | NM_001168.2 | Refseq |
| Affymetrix Human Clariom D Assay | TC1700009043.hg.1 | NM_001168.2 | Refseq |
| Affymetrix Human Clariom D Assay | TC1700009043.hg.1 | NM_001168.2 | Refseq |
| Affymetrix Human Clariom D Assay | TC1700009043.hg.1 | NM_001168.2 | Refseq |
| Affymetrix Human Clariom D Assay | TC1700009043.hg.1 | NM_001168.2 | Refseq |

BIRC5
Genomic Map