Assay Details
| Gene Symbol: EXD1 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | NM_001286441.1 | NP_001273370.1 | 8 - 9 | 972 | 92 | |
| NM_152596.3 | NP_689809.2 | 6 - 7 | 717 | 92 | ||
| XM_006720415.3 | XP_006720478.1 | 7 - 8 | 889 | 92 | ||
| XM_011521298.2 | XP_011519600.1 | 9 - 10 | 1202 | 92 | ||
| XM_011521299.2 | XP_011519601.1 | 9 - 10 | 933 | 92 | ||
| XM_011521300.2 | XP_011519602.1 | 6 - 7 | 461 | 92 | ||
| XM_011521301.2 | XP_011519603.1 | 8 - 9 | 758 | 92 | ||
| XM_011521302.2 | XP_011519604.1 | 3 - 4 | 275 | 92 | ||
| XM_017021970.1 | XP_016877459.1 | 6 - 7 | 461 | 92 | ||
| XM_017021971.1 | XP_016877460.1 | 6 - 7 | 459 | 92 | ||
| GenBank mRNA | AK292524.1 | - | 6 - 7 | 623 | 92 | |
| AK302864.1 | - | 8 - 9 | 906 | 92 | ||
| BC030628.2 | - | 6 - 7 | 657 | 4827307 | 92 | |
More Information
Updated Mapping Information
This assay targets the same location as HTA transcript cluster(s) TC15001247.hg.1
Additional Information:
This assay is in the 5'UTR region of the XM_011521302.2 transcripts.
Set Membership: |
|
Matching Ambion siRNAs:
Gene Ontology Categories:
MicroArray Probes:
Affymetrix
| Array Name | Probe IDs | Targets | Target Type |
|---|---|---|---|
| Affymetrix Human Transcriptome Array 2.0 | PSR15011314.hg.1 | ENST00000314992.5 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | PSR15011314.hg.1 | ENST00000458580.2 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | PSR15011314.hg.1 | NM_152596.3 | Refseq |
| Affymetrix Human Transcriptome Array 2.0 | PSR15011314.hg.1 | TC15001247.hg.1 | HTA TC |
| Affymetrix Human Transcriptome Array 2.0 | TC1500009143.hg.1 | ENST00000314992.9 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC1500009143.hg.1 | ENST00000314992.9 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC1500009143.hg.1 | ENST00000314992.9 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC1500009143.hg.1 | ENST00000314992.9 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC1500009143.hg.1 | ENST00000314992.9 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC1500009143.hg.1 | ENST00000314992.9 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC1500009143.hg.1 | ENST00000314992.9 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC1500009143.hg.1 | ENST00000314992.9 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC1500009143.hg.1 | ENST00000314992.9 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC1500009143.hg.1 | ENST00000314992.9 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC1500009143.hg.1 | ENST00000314992.9 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC1500009143.hg.1 | ENST00000314992.9 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC1500009143.hg.1 | ENST00000314992.9 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC1500009143.hg.1 | ENST00000314992.9 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC1500009143.hg.1 | ENST00000314992.9 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC1500009143.hg.1 | ENST00000314992.9 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC1500009143.hg.1 | ENST00000458580.6 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC1500009143.hg.1 | ENST00000458580.6 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC1500009143.hg.1 | ENST00000458580.6 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC1500009143.hg.1 | ENST00000458580.6 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC1500009143.hg.1 | ENST00000458580.6 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC1500009143.hg.1 | ENST00000458580.6 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC1500009143.hg.1 | ENST00000458580.6 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC1500009143.hg.1 | ENST00000458580.6 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC1500009143.hg.1 | ENST00000458580.6 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC1500009143.hg.1 | ENST00000458580.6 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC1500009143.hg.1 | ENST00000458580.6 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC1500009143.hg.1 | ENST00000458580.6 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC1500009143.hg.1 | ENST00000458580.6 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC1500009143.hg.1 | ENST00000458580.6 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC1500009143.hg.1 | ENST00000458580.6 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC1500009143.hg.1 | ENST00000458580.6 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC1500009143.hg.1 | ENST00000458580.6 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC1500009143.hg.1 | ENST00000458580.6 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC1500009143.hg.1 | ENST00000558396.1 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC1500009143.hg.1 | ENST00000558396.1 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC1500009143.hg.1 | ENST00000558396.1 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC1500009143.hg.1 | ENST00000558396.1 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC1500009143.hg.1 | ENST00000558396.1 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC1500009143.hg.1 | ENST00000558396.1 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC1500009143.hg.1 | ENST00000558396.1 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC1500009143.hg.1 | ENST00000558881.1 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC1500009143.hg.1 | ENST00000558881.1 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC1500009143.hg.1 | ENST00000558881.1 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC1500009143.hg.1 | ENST00000558881.1 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC1500009143.hg.1 | ENST00000558881.1 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC1500009143.hg.1 | ENST00000559743.1 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC1500009143.hg.1 | ENST00000559743.1 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC1500009143.hg.1 | ENST00000559743.1 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC1500009143.hg.1 | ENST00000559743.1 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC1500009143.hg.1 | ENST00000559743.1 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC1500009143.hg.1 | ENST00000559743.1 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC1500009143.hg.1 | ENST00000559743.1 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC1500009143.hg.1 | ENST00000559743.1 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC1500009143.hg.1 | ENST00000559743.1 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC1500009143.hg.1 | NM_001286441.1 | Refseq |
| Affymetrix Human Transcriptome Array 2.0 | TC1500009143.hg.1 | NM_001286441.1 | Refseq |
| Affymetrix Human Transcriptome Array 2.0 | TC1500009143.hg.1 | NM_001286441.1 | Refseq |
| Affymetrix Human Transcriptome Array 2.0 | TC1500009143.hg.1 | NM_001286441.1 | Refseq |
| Affymetrix Human Transcriptome Array 2.0 | TC1500009143.hg.1 | NM_001286441.1 | Refseq |
| Affymetrix Human Transcriptome Array 2.0 | TC1500009143.hg.1 | NM_001286441.1 | Refseq |
| Affymetrix Human Transcriptome Array 2.0 | TC1500009143.hg.1 | NM_001286441.1 | Refseq |
| Affymetrix Human Transcriptome Array 2.0 | TC1500009143.hg.1 | NM_001286441.1 | Refseq |
| Affymetrix Human Transcriptome Array 2.0 | TC1500009143.hg.1 | NM_001286441.1 | Refseq |
| Affymetrix Human Transcriptome Array 2.0 | TC1500009143.hg.1 | NM_001286441.1 | Refseq |
| Affymetrix Human Transcriptome Array 2.0 | TC1500009143.hg.1 | NM_001286441.1 | Refseq |
| Affymetrix Human Transcriptome Array 2.0 | TC1500009143.hg.1 | NM_001286441.1 | Refseq |
| Affymetrix Human Transcriptome Array 2.0 | TC1500009143.hg.1 | NM_001286441.1 | Refseq |
| Affymetrix Human Transcriptome Array 2.0 | TC1500009143.hg.1 | NM_001286441.1 | Refseq |
| Affymetrix Human Transcriptome Array 2.0 | TC1500009143.hg.1 | NM_001286441.1 | Refseq |
| Affymetrix Human Transcriptome Array 2.0 | TC1500009143.hg.1 | NM_001286441.1 | Refseq |
| Affymetrix Human Transcriptome Array 2.0 | TC1500009143.hg.1 | NM_001286441.1 | Refseq |
| Affymetrix Human Transcriptome Array 2.0 | TC1500009143.hg.1 | NM_001286441.1 | Refseq |
| Affymetrix Human Transcriptome Array 2.0 | TC1500009143.hg.1 | NM_001286441.1 | Refseq |
| Affymetrix Human Transcriptome Array 2.0 | TC1500009143.hg.1 | NM_001286441.1 | Refseq |
| Affymetrix Human Transcriptome Array 2.0 | TC1500009143.hg.1 | NM_152596.3 | Refseq |
| Affymetrix Human Transcriptome Array 2.0 | TC1500009143.hg.1 | NM_152596.3 | Refseq |
| Affymetrix Human Transcriptome Array 2.0 | TC1500009143.hg.1 | NM_152596.3 | Refseq |
| Affymetrix Human Transcriptome Array 2.0 | TC1500009143.hg.1 | NM_152596.3 | Refseq |
| Affymetrix Human Transcriptome Array 2.0 | TC1500009143.hg.1 | NM_152596.3 | Refseq |
| Affymetrix Human Transcriptome Array 2.0 | TC1500009143.hg.1 | NM_152596.3 | Refseq |
| Affymetrix Human Transcriptome Array 2.0 | TC1500009143.hg.1 | NM_152596.3 | Refseq |
| Affymetrix Human Transcriptome Array 2.0 | TC1500009143.hg.1 | NM_152596.3 | Refseq |
| Affymetrix Human Transcriptome Array 2.0 | TC1500009143.hg.1 | NM_152596.3 | Refseq |
| Affymetrix Human Transcriptome Array 2.0 | TC1500009143.hg.1 | NM_152596.3 | Refseq |
| Affymetrix Human Transcriptome Array 2.0 | TC1500009143.hg.1 | NM_152596.3 | Refseq |
| Affymetrix Human Transcriptome Array 2.0 | TC1500009143.hg.1 | NM_152596.3 | Refseq |
| Affymetrix Human Transcriptome Array 2.0 | TC1500009143.hg.1 | NM_152596.3 | Refseq |
| Affymetrix Human Transcriptome Array 2.0 | TC1500009143.hg.1 | NM_152596.3 | Refseq |
| Affymetrix Human Transcriptome Array 2.0 | TC1500009143.hg.1 | NM_152596.3 | Refseq |
| Affymetrix Human Transcriptome Array 2.0 | TC1500009143.hg.1 | NM_152596.3 | Refseq |
| Affymetrix Human Transcriptome Array 2.0 | TC1500009143.hg.1 | NM_152596.3 | Refseq |
| Affymetrix Human Clariom D Assay | PSR1500166233.hg.1 | ENST00000314992.9 | Ensembl |
| Affymetrix Human Clariom D Assay | PSR1500166233.hg.1 | ENST00000458580.6 | Ensembl |
| Affymetrix Human Clariom D Assay | PSR1500166233.hg.1 | NM_001286441.1 | Refseq |
| Affymetrix Human Clariom D Assay | PSR1500166233.hg.1 | NM_152596.3 | Refseq |
| Affymetrix Human Clariom D Assay | PSR1500166233.hg.1 | ENST00000314992.9 | Ensembl |
| Affymetrix Human Clariom D Assay | PSR1500166233.hg.1 | ENST00000458580.6 | Ensembl |
| Affymetrix Human Clariom D Assay | PSR1500166233.hg.1 | NM_001286441.1 | Refseq |
| Affymetrix Human Clariom D Assay | PSR1500166233.hg.1 | NM_152596.3 | Refseq |
| Affymetrix Human Clariom D Assay | PSR1500166233.hg.1 | TC1500009143.hg.1 | Clariom D TC |
| Affymetrix Human Clariom D Assay | TC1500009143.hg.1 | ENST00000314992.9 | Ensembl |
| Affymetrix Human Clariom D Assay | TC1500009143.hg.1 | ENST00000314992.9 | Ensembl |
| Affymetrix Human Clariom D Assay | TC1500009143.hg.1 | ENST00000314992.9 | Ensembl |
| Affymetrix Human Clariom D Assay | TC1500009143.hg.1 | ENST00000314992.9 | Ensembl |
| Affymetrix Human Clariom D Assay | TC1500009143.hg.1 | ENST00000314992.9 | Ensembl |
| Affymetrix Human Clariom D Assay | TC1500009143.hg.1 | ENST00000314992.9 | Ensembl |
| Affymetrix Human Clariom D Assay | TC1500009143.hg.1 | ENST00000314992.9 | Ensembl |
| Affymetrix Human Clariom D Assay | TC1500009143.hg.1 | ENST00000314992.9 | Ensembl |
| Affymetrix Human Clariom D Assay | TC1500009143.hg.1 | ENST00000314992.9 | Ensembl |
| Affymetrix Human Clariom D Assay | TC1500009143.hg.1 | ENST00000314992.9 | Ensembl |
| Affymetrix Human Clariom D Assay | TC1500009143.hg.1 | ENST00000314992.9 | Ensembl |
| Affymetrix Human Clariom D Assay | TC1500009143.hg.1 | ENST00000314992.9 | Ensembl |
| Affymetrix Human Clariom D Assay | TC1500009143.hg.1 | ENST00000314992.9 | Ensembl |
| Affymetrix Human Clariom D Assay | TC1500009143.hg.1 | ENST00000314992.9 | Ensembl |
| Affymetrix Human Clariom D Assay | TC1500009143.hg.1 | ENST00000314992.9 | Ensembl |
| Affymetrix Human Clariom D Assay | TC1500009143.hg.1 | ENST00000314992.9 | Ensembl |
| Affymetrix Human Clariom D Assay | TC1500009143.hg.1 | ENST00000458580.6 | Ensembl |
| Affymetrix Human Clariom D Assay | TC1500009143.hg.1 | ENST00000458580.6 | Ensembl |
| Affymetrix Human Clariom D Assay | TC1500009143.hg.1 | ENST00000458580.6 | Ensembl |
| Affymetrix Human Clariom D Assay | TC1500009143.hg.1 | ENST00000458580.6 | Ensembl |
| Affymetrix Human Clariom D Assay | TC1500009143.hg.1 | ENST00000458580.6 | Ensembl |
| Affymetrix Human Clariom D Assay | TC1500009143.hg.1 | ENST00000458580.6 | Ensembl |
| Affymetrix Human Clariom D Assay | TC1500009143.hg.1 | ENST00000458580.6 | Ensembl |
| Affymetrix Human Clariom D Assay | TC1500009143.hg.1 | ENST00000458580.6 | Ensembl |
| Affymetrix Human Clariom D Assay | TC1500009143.hg.1 | ENST00000458580.6 | Ensembl |
| Affymetrix Human Clariom D Assay | TC1500009143.hg.1 | ENST00000458580.6 | Ensembl |
| Affymetrix Human Clariom D Assay | TC1500009143.hg.1 | ENST00000458580.6 | Ensembl |
| Affymetrix Human Clariom D Assay | TC1500009143.hg.1 | ENST00000458580.6 | Ensembl |
| Affymetrix Human Clariom D Assay | TC1500009143.hg.1 | ENST00000458580.6 | Ensembl |
| Affymetrix Human Clariom D Assay | TC1500009143.hg.1 | ENST00000458580.6 | Ensembl |
| Affymetrix Human Clariom D Assay | TC1500009143.hg.1 | ENST00000458580.6 | Ensembl |
| Affymetrix Human Clariom D Assay | TC1500009143.hg.1 | ENST00000458580.6 | Ensembl |
| Affymetrix Human Clariom D Assay | TC1500009143.hg.1 | ENST00000458580.6 | Ensembl |
| Affymetrix Human Clariom D Assay | TC1500009143.hg.1 | ENST00000458580.6 | Ensembl |
| Affymetrix Human Clariom D Assay | TC1500009143.hg.1 | ENST00000558396.1 | Ensembl |
| Affymetrix Human Clariom D Assay | TC1500009143.hg.1 | ENST00000558396.1 | Ensembl |
| Affymetrix Human Clariom D Assay | TC1500009143.hg.1 | ENST00000558396.1 | Ensembl |
| Affymetrix Human Clariom D Assay | TC1500009143.hg.1 | ENST00000558396.1 | Ensembl |
| Affymetrix Human Clariom D Assay | TC1500009143.hg.1 | ENST00000558396.1 | Ensembl |
| Affymetrix Human Clariom D Assay | TC1500009143.hg.1 | ENST00000558396.1 | Ensembl |
| Affymetrix Human Clariom D Assay | TC1500009143.hg.1 | ENST00000558396.1 | Ensembl |
| Affymetrix Human Clariom D Assay | TC1500009143.hg.1 | ENST00000558881.1 | Ensembl |
| Affymetrix Human Clariom D Assay | TC1500009143.hg.1 | ENST00000558881.1 | Ensembl |
| Affymetrix Human Clariom D Assay | TC1500009143.hg.1 | ENST00000558881.1 | Ensembl |
| Affymetrix Human Clariom D Assay | TC1500009143.hg.1 | ENST00000558881.1 | Ensembl |
| Affymetrix Human Clariom D Assay | TC1500009143.hg.1 | ENST00000558881.1 | Ensembl |
| Affymetrix Human Clariom D Assay | TC1500009143.hg.1 | ENST00000559743.1 | Ensembl |
| Affymetrix Human Clariom D Assay | TC1500009143.hg.1 | ENST00000559743.1 | Ensembl |
| Affymetrix Human Clariom D Assay | TC1500009143.hg.1 | ENST00000559743.1 | Ensembl |
| Affymetrix Human Clariom D Assay | TC1500009143.hg.1 | ENST00000559743.1 | Ensembl |
| Affymetrix Human Clariom D Assay | TC1500009143.hg.1 | ENST00000559743.1 | Ensembl |
| Affymetrix Human Clariom D Assay | TC1500009143.hg.1 | ENST00000559743.1 | Ensembl |
| Affymetrix Human Clariom D Assay | TC1500009143.hg.1 | ENST00000559743.1 | Ensembl |
| Affymetrix Human Clariom D Assay | TC1500009143.hg.1 | ENST00000559743.1 | Ensembl |
| Affymetrix Human Clariom D Assay | TC1500009143.hg.1 | ENST00000559743.1 | Ensembl |
| Affymetrix Human Clariom D Assay | TC1500009143.hg.1 | NM_001286441.1 | Refseq |
| Affymetrix Human Clariom D Assay | TC1500009143.hg.1 | NM_001286441.1 | Refseq |
| Affymetrix Human Clariom D Assay | TC1500009143.hg.1 | NM_001286441.1 | Refseq |
| Affymetrix Human Clariom D Assay | TC1500009143.hg.1 | NM_001286441.1 | Refseq |
| Affymetrix Human Clariom D Assay | TC1500009143.hg.1 | NM_001286441.1 | Refseq |
| Affymetrix Human Clariom D Assay | TC1500009143.hg.1 | NM_001286441.1 | Refseq |
| Affymetrix Human Clariom D Assay | TC1500009143.hg.1 | NM_001286441.1 | Refseq |
| Affymetrix Human Clariom D Assay | TC1500009143.hg.1 | NM_001286441.1 | Refseq |
| Affymetrix Human Clariom D Assay | TC1500009143.hg.1 | NM_001286441.1 | Refseq |
| Affymetrix Human Clariom D Assay | TC1500009143.hg.1 | NM_001286441.1 | Refseq |
| Affymetrix Human Clariom D Assay | TC1500009143.hg.1 | NM_001286441.1 | Refseq |
| Affymetrix Human Clariom D Assay | TC1500009143.hg.1 | NM_001286441.1 | Refseq |
| Affymetrix Human Clariom D Assay | TC1500009143.hg.1 | NM_001286441.1 | Refseq |
| Affymetrix Human Clariom D Assay | TC1500009143.hg.1 | NM_001286441.1 | Refseq |
| Affymetrix Human Clariom D Assay | TC1500009143.hg.1 | NM_001286441.1 | Refseq |
| Affymetrix Human Clariom D Assay | TC1500009143.hg.1 | NM_001286441.1 | Refseq |
| Affymetrix Human Clariom D Assay | TC1500009143.hg.1 | NM_001286441.1 | Refseq |
| Affymetrix Human Clariom D Assay | TC1500009143.hg.1 | NM_001286441.1 | Refseq |
| Affymetrix Human Clariom D Assay | TC1500009143.hg.1 | NM_001286441.1 | Refseq |
| Affymetrix Human Clariom D Assay | TC1500009143.hg.1 | NM_001286441.1 | Refseq |
| Affymetrix Human Clariom D Assay | TC1500009143.hg.1 | NM_152596.3 | Refseq |
| Affymetrix Human Clariom D Assay | TC1500009143.hg.1 | NM_152596.3 | Refseq |
| Affymetrix Human Clariom D Assay | TC1500009143.hg.1 | NM_152596.3 | Refseq |
| Affymetrix Human Clariom D Assay | TC1500009143.hg.1 | NM_152596.3 | Refseq |
| Affymetrix Human Clariom D Assay | TC1500009143.hg.1 | NM_152596.3 | Refseq |
| Affymetrix Human Clariom D Assay | TC1500009143.hg.1 | NM_152596.3 | Refseq |
| Affymetrix Human Clariom D Assay | TC1500009143.hg.1 | NM_152596.3 | Refseq |
| Affymetrix Human Clariom D Assay | TC1500009143.hg.1 | NM_152596.3 | Refseq |
| Affymetrix Human Clariom D Assay | TC1500009143.hg.1 | NM_152596.3 | Refseq |
| Affymetrix Human Clariom D Assay | TC1500009143.hg.1 | NM_152596.3 | Refseq |
| Affymetrix Human Clariom D Assay | TC1500009143.hg.1 | NM_152596.3 | Refseq |
| Affymetrix Human Clariom D Assay | TC1500009143.hg.1 | NM_152596.3 | Refseq |
| Affymetrix Human Clariom D Assay | TC1500009143.hg.1 | NM_152596.3 | Refseq |
| Affymetrix Human Clariom D Assay | TC1500009143.hg.1 | NM_152596.3 | Refseq |
| Affymetrix Human Clariom D Assay | TC1500009143.hg.1 | NM_152596.3 | Refseq |
| Affymetrix Human Clariom D Assay | TC1500009143.hg.1 | NM_152596.3 | Refseq |
| Affymetrix Human Clariom D Assay | TC1500009143.hg.1 | NM_152596.3 | Refseq |

EXD1
Genomic Map