Assay Details
| Gene Symbol: PITPNB | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | NM_001284277.1 | NP_001271206.1 | 2 - 3 | 161 | 96 | |
| NM_012399.4 | NP_036531.1 | 2 - 3 | 161 | 96 | ||
| GenBank mRNA | AK313064.1 | - | 2 - 3 | 111 | 96 | |
| BC018704.1 | - | 2 - 3 | 85 | 4643509 | 96 | |
| BC031427.1 | - | 2 - 3 | 78 | 4341137 | 96 | |
| BU147254.1 | - | 2 - 3 | 122 | 96 | ||
| CR456541.1 | - | 2 - 3 | 113 | 96 | ||
| D30037.1 | - | 2 - 3 | 92 | 96 | ||
| HY176246.1 | - | 2 - 3 | 161 | 96 | ||
More Information
Updated Mapping Information
Set Membership: |
|

PITPNB
Genomic Map