Assay Details
| Gene Symbol: TTL | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | NM_153712.4 | NP_714923.1 | 1 - 2 | 334 | 84 | |
| XM_005263599.3 | XP_005263656.1 | 1 - 2 | 373 | 84 | ||
| XM_011510665.2 | XP_011508967.1 | 1 - 2 | 373 | 84 | ||
| GenBank mRNA | AB071393.2 | - | 1 - 2 | 179 | 84 | |
| BC036819.1 | - | 1 - 2 | 346 | 5767758 | 84 | |
More Information
Updated Mapping Information
This assay targets the same location as HTA transcript cluster(s) TC02000706.hg.1
Set Membership: |
|
Matching Ambion siRNAs:
Gene Ontology Categories:
MicroArray Probes:
Affymetrix
ABI
| Array Name | Probe IDs | Targets | Target Type |
|---|---|---|---|
| Affymetrix Human Transcriptome Array 2.0 | PSR02010916.hg.1 | TC02000706.hg.1 | HTA TC |
| Affymetrix Human Transcriptome Array 2.0 | PSR02010916.hg.1 | ENST00000233336.5 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | PSR02010916.hg.1 | NM_153712.4 | Refseq |
| Affymetrix Human Transcriptome Array 2.0 | PSR02010917.hg.1 | TC02000706.hg.1 | HTA TC |
| Affymetrix Human Transcriptome Array 2.0 | PSR02010917.hg.1 | ENST00000233336.5 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | PSR02010917.hg.1 | NM_153712.4 | Refseq |
| Affymetrix Human Transcriptome Array 2.0 | PSR02010919.hg.1 | TC02000706.hg.1 | HTA TC |
| Affymetrix Human Transcriptome Array 2.0 | PSR02010919.hg.1 | ENST00000233336.5 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | PSR02010919.hg.1 | NM_153712.4 | Refseq |
| Affymetrix Human Transcriptome Array 2.0 | TC0200008921.hg.1 | ENST00000233336.6 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0200008921.hg.1 | ENST00000233336.6 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0200008921.hg.1 | ENST00000233336.6 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0200008921.hg.1 | ENST00000233336.6 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0200008921.hg.1 | ENST00000233336.6 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0200008921.hg.1 | ENST00000233336.6 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0200008921.hg.1 | ENST00000233336.6 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0200008921.hg.1 | ENST00000233336.6 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0200008921.hg.1 | ENST00000233336.6 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0200008921.hg.1 | ENST00000233336.6 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0200008921.hg.1 | ENST00000233336.6 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0200008921.hg.1 | ENST00000233336.6 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0200008921.hg.1 | ENST00000233336.6 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0200008921.hg.1 | ENST00000233336.6 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0200008921.hg.1 | ENST00000460450.1 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0200008921.hg.1 | ENST00000460450.1 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0200008921.hg.1 | ENST00000460450.1 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0200008921.hg.1 | ENST00000460450.1 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0200008921.hg.1 | ENST00000476615.1 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0200008921.hg.1 | ENST00000476615.1 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0200008921.hg.1 | ENST00000476615.1 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0200008921.hg.1 | ENST00000476615.1 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0200008921.hg.1 | ENST00000485537.1 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0200008921.hg.1 | ENST00000485537.1 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0200008921.hg.1 | ENST00000485537.1 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0200008921.hg.1 | NM_153712.4 | Refseq |
| Affymetrix Human Transcriptome Array 2.0 | TC0200008921.hg.1 | NM_153712.4 | Refseq |
| Affymetrix Human Transcriptome Array 2.0 | TC0200008921.hg.1 | NM_153712.4 | Refseq |
| Affymetrix Human Transcriptome Array 2.0 | TC0200008921.hg.1 | NM_153712.4 | Refseq |
| Affymetrix Human Transcriptome Array 2.0 | TC0200008921.hg.1 | NM_153712.4 | Refseq |
| Affymetrix Human Transcriptome Array 2.0 | TC0200008921.hg.1 | NM_153712.4 | Refseq |
| Affymetrix Human Transcriptome Array 2.0 | TC0200008921.hg.1 | NM_153712.4 | Refseq |
| Affymetrix Human Transcriptome Array 2.0 | TC0200008921.hg.1 | NM_153712.4 | Refseq |
| Affymetrix Human Transcriptome Array 2.0 | TC0200008921.hg.1 | NM_153712.4 | Refseq |
| Affymetrix Human Transcriptome Array 2.0 | TC0200008921.hg.1 | NM_153712.4 | Refseq |
| Affymetrix Human Transcriptome Array 2.0 | TC0200008921.hg.1 | NM_153712.4 | Refseq |
| Affymetrix Human Transcriptome Array 2.0 | TC0200008921.hg.1 | NM_153712.4 | Refseq |
| Affymetrix Human Transcriptome Array 2.0 | TC0200008921.hg.1 | NM_153712.4 | Refseq |
| Affymetrix Human Clariom D Assay | PSR0200162244.hg.1 | ENST00000233336.6 | Ensembl |
| Affymetrix Human Clariom D Assay | PSR0200162244.hg.1 | ENST00000485537.1 | Ensembl |
| Affymetrix Human Clariom D Assay | PSR0200162244.hg.1 | NM_153712.4 | Refseq |
| Affymetrix Human Clariom D Assay | PSR0200162244.hg.1 | ENST00000233336.6 | Ensembl |
| Affymetrix Human Clariom D Assay | PSR0200162244.hg.1 | ENST00000485537.1 | Ensembl |
| Affymetrix Human Clariom D Assay | PSR0200162244.hg.1 | NM_153712.4 | Refseq |
| Affymetrix Human Clariom D Assay | PSR0200162244.hg.1 | TC0200008921.hg.1 | Clariom D TC |
| Affymetrix Human Clariom D Assay | PSR0200162247.hg.1 | ENST00000233336.6 | Ensembl |
| Affymetrix Human Clariom D Assay | PSR0200162247.hg.1 | NM_153712.4 | Refseq |
| Affymetrix Human Clariom D Assay | PSR0200162247.hg.1 | ENST00000233336.6 | Ensembl |
| Affymetrix Human Clariom D Assay | PSR0200162247.hg.1 | NM_153712.4 | Refseq |
| Affymetrix Human Clariom D Assay | PSR0200162247.hg.1 | TC0200008921.hg.1 | Clariom D TC |
| Affymetrix Human Clariom D Assay | TC0200008921.hg.1 | ENST00000233336.6 | Ensembl |
| Affymetrix Human Clariom D Assay | TC0200008921.hg.1 | ENST00000233336.6 | Ensembl |
| Affymetrix Human Clariom D Assay | TC0200008921.hg.1 | ENST00000233336.6 | Ensembl |
| Affymetrix Human Clariom D Assay | TC0200008921.hg.1 | ENST00000233336.6 | Ensembl |
| Affymetrix Human Clariom D Assay | TC0200008921.hg.1 | ENST00000233336.6 | Ensembl |
| Affymetrix Human Clariom D Assay | TC0200008921.hg.1 | ENST00000233336.6 | Ensembl |
| Affymetrix Human Clariom D Assay | TC0200008921.hg.1 | ENST00000233336.6 | Ensembl |
| Affymetrix Human Clariom D Assay | TC0200008921.hg.1 | ENST00000233336.6 | Ensembl |
| Affymetrix Human Clariom D Assay | TC0200008921.hg.1 | ENST00000233336.6 | Ensembl |
| Affymetrix Human Clariom D Assay | TC0200008921.hg.1 | ENST00000233336.6 | Ensembl |
| Affymetrix Human Clariom D Assay | TC0200008921.hg.1 | ENST00000233336.6 | Ensembl |
| Affymetrix Human Clariom D Assay | TC0200008921.hg.1 | ENST00000233336.6 | Ensembl |
| Affymetrix Human Clariom D Assay | TC0200008921.hg.1 | ENST00000233336.6 | Ensembl |
| Affymetrix Human Clariom D Assay | TC0200008921.hg.1 | ENST00000233336.6 | Ensembl |
| Affymetrix Human Clariom D Assay | TC0200008921.hg.1 | ENST00000460450.1 | Ensembl |
| Affymetrix Human Clariom D Assay | TC0200008921.hg.1 | ENST00000460450.1 | Ensembl |
| Affymetrix Human Clariom D Assay | TC0200008921.hg.1 | ENST00000460450.1 | Ensembl |
| Affymetrix Human Clariom D Assay | TC0200008921.hg.1 | ENST00000460450.1 | Ensembl |
| Affymetrix Human Clariom D Assay | TC0200008921.hg.1 | ENST00000476615.1 | Ensembl |
| Affymetrix Human Clariom D Assay | TC0200008921.hg.1 | ENST00000476615.1 | Ensembl |
| Affymetrix Human Clariom D Assay | TC0200008921.hg.1 | ENST00000476615.1 | Ensembl |
| Affymetrix Human Clariom D Assay | TC0200008921.hg.1 | ENST00000476615.1 | Ensembl |
| Affymetrix Human Clariom D Assay | TC0200008921.hg.1 | ENST00000485537.1 | Ensembl |
| Affymetrix Human Clariom D Assay | TC0200008921.hg.1 | ENST00000485537.1 | Ensembl |
| Affymetrix Human Clariom D Assay | TC0200008921.hg.1 | ENST00000485537.1 | Ensembl |
| Affymetrix Human Clariom D Assay | TC0200008921.hg.1 | NM_153712.4 | Refseq |
| Affymetrix Human Clariom D Assay | TC0200008921.hg.1 | NM_153712.4 | Refseq |
| Affymetrix Human Clariom D Assay | TC0200008921.hg.1 | NM_153712.4 | Refseq |
| Affymetrix Human Clariom D Assay | TC0200008921.hg.1 | NM_153712.4 | Refseq |
| Affymetrix Human Clariom D Assay | TC0200008921.hg.1 | NM_153712.4 | Refseq |
| Affymetrix Human Clariom D Assay | TC0200008921.hg.1 | NM_153712.4 | Refseq |
| Affymetrix Human Clariom D Assay | TC0200008921.hg.1 | NM_153712.4 | Refseq |
| Affymetrix Human Clariom D Assay | TC0200008921.hg.1 | NM_153712.4 | Refseq |
| Affymetrix Human Clariom D Assay | TC0200008921.hg.1 | NM_153712.4 | Refseq |
| Affymetrix Human Clariom D Assay | TC0200008921.hg.1 | NM_153712.4 | Refseq |
| Affymetrix Human Clariom D Assay | TC0200008921.hg.1 | NM_153712.4 | Refseq |
| Affymetrix Human Clariom D Assay | TC0200008921.hg.1 | NM_153712.4 | Refseq |
| Affymetrix Human Clariom D Assay | TC0200008921.hg.1 | NM_153712.4 | Refseq |
| Affymetrix Human Clariom D Assay | PSR0200162243.hg.1 | ENST00000233336.6 | Ensembl |
| Affymetrix Human Clariom D Assay | PSR0200162243.hg.1 | NM_153712.4 | Refseq |
| Affymetrix Human Clariom D Assay | PSR0200162243.hg.1 | ENST00000233336.6 | Ensembl |
| Affymetrix Human Clariom D Assay | PSR0200162243.hg.1 | NM_153712.4 | Refseq |
| Affymetrix Human Clariom D Assay | PSR0200162243.hg.1 | TC0200008921.hg.1 | Clariom D TC |
| Affymetrix Human Clariom D Assay | PSR0200162245.hg.1 | ENST00000233336.6 | Ensembl |
| Affymetrix Human Clariom D Assay | PSR0200162245.hg.1 | ENST00000485537.1 | Ensembl |
| Affymetrix Human Clariom D Assay | PSR0200162245.hg.1 | NM_153712.4 | Refseq |
| Affymetrix Human Clariom D Assay | PSR0200162245.hg.1 | ENST00000233336.6 | Ensembl |
| Affymetrix Human Clariom D Assay | PSR0200162245.hg.1 | ENST00000485537.1 | Ensembl |
| Affymetrix Human Clariom D Assay | PSR0200162245.hg.1 | NM_153712.4 | Refseq |
| Affymetrix Human Clariom D Assay | PSR0200162245.hg.1 | TC0200008921.hg.1 | Clariom D TC |
| Array Name | Probe IDs | Targets | Target Type |
|---|---|---|---|
| ABI Human Genome Survey Microarray Version 2.0 | 223304 | - | - |

TTL
Genomic Map