Assay Details
| Gene Symbol: IFNGR2 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | NM_005534.3 | NP_005525.2 | 5 - 6 | 1365 | 83 | |
| XM_005260969.2 | XP_005261026.1 | 6 - 7 | 881 | 83 | ||
| XM_011529553.1 | XP_011527855.1 | 6 - 7 | 807 | 83 | ||
| XM_011529554.2 | XP_011527856.1 | 5 - 6 | 1241 | 83 | ||
| GenBank mRNA | AK292246.1 | - | 5 - 6 | 813 | 83 | |
| AK314005.1 | - | 5 - 6 | 826 | 83 | ||
| BC003624.2 | - | 5 - 6 | 833 | 2967074 | 83 | |
| U05875.1 | - | 5 - 6 | 1365 | 83 | ||
| U05877.1 | - | 5 - 6 | 830 | 83 | ||
More Information
Updated Mapping Information
This assay targets the same location as HTA transcript cluster(s) TC21000129.hg.1
Set Membership: |
|
Matching Ambion siRNAs:
Panther Classification:
Gene Ontology Categories:
MicroArray Probes:
Affymetrix
| Array Name | Probe IDs | Targets | Target Type |
|---|---|---|---|
| Affymetrix Human Transcriptome Array 2.0 | TC2100006967.hg.1 | ENST00000290219.10 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC2100006967.hg.1 | ENST00000290219.10 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC2100006967.hg.1 | ENST00000290219.10 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC2100006967.hg.1 | ENST00000290219.10 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC2100006967.hg.1 | ENST00000290219.10 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC2100006967.hg.1 | ENST00000290219.10 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC2100006967.hg.1 | ENST00000290219.10 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC2100006967.hg.1 | ENST00000290219.10 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC2100006967.hg.1 | ENST00000290219.10 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC2100006967.hg.1 | ENST00000290219.10 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC2100006967.hg.1 | ENST00000290219.10 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC2100006967.hg.1 | ENST00000290219.10 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC2100006967.hg.1 | ENST00000381995.5 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC2100006967.hg.1 | ENST00000381995.5 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC2100006967.hg.1 | ENST00000381995.5 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC2100006967.hg.1 | ENST00000381995.5 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC2100006967.hg.1 | ENST00000381995.5 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC2100006967.hg.1 | ENST00000381995.5 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC2100006967.hg.1 | ENST00000381995.5 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC2100006967.hg.1 | ENST00000381995.5 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC2100006967.hg.1 | ENST00000381995.5 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC2100006967.hg.1 | ENST00000381995.5 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC2100006967.hg.1 | ENST00000381995.5 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC2100006967.hg.1 | ENST00000381995.5 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC2100006967.hg.1 | ENST00000405436.5 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC2100006967.hg.1 | ENST00000405436.5 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC2100006967.hg.1 | ENST00000405436.5 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC2100006967.hg.1 | ENST00000405436.5 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC2100006967.hg.1 | ENST00000405436.5 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC2100006967.hg.1 | ENST00000405436.5 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC2100006967.hg.1 | ENST00000405436.5 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC2100006967.hg.1 | ENST00000405436.5 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC2100006967.hg.1 | ENST00000405436.5 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC2100006967.hg.1 | ENST00000405436.5 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC2100006967.hg.1 | ENST00000405436.5 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC2100006967.hg.1 | ENST00000421802.1 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC2100006967.hg.1 | ENST00000421802.1 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC2100006967.hg.1 | ENST00000439213.5 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC2100006967.hg.1 | ENST00000439213.5 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC2100006967.hg.1 | ENST00000439213.5 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC2100006967.hg.1 | ENST00000439213.5 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC2100006967.hg.1 | ENST00000439213.5 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC2100006967.hg.1 | ENST00000439213.5 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC2100006967.hg.1 | ENST00000439213.5 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC2100006967.hg.1 | ENST00000439213.5 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC2100006967.hg.1 | ENST00000439213.5 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC2100006967.hg.1 | ENST00000545369.2 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC2100006967.hg.1 | ENST00000545369.2 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC2100006967.hg.1 | ENST00000545369.2 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC2100006967.hg.1 | ENST00000545369.2 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC2100006967.hg.1 | ENST00000545369.2 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC2100006967.hg.1 | ENST00000545369.2 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC2100006967.hg.1 | ENST00000545369.2 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC2100006967.hg.1 | ENST00000545369.2 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC2100006967.hg.1 | ENST00000545369.2 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC2100006967.hg.1 | NM_005534.3 | Refseq |
| Affymetrix Human Transcriptome Array 2.0 | TC2100006967.hg.1 | NM_005534.3 | Refseq |
| Affymetrix Human Transcriptome Array 2.0 | TC2100006967.hg.1 | NM_005534.3 | Refseq |
| Affymetrix Human Transcriptome Array 2.0 | TC2100006967.hg.1 | NM_005534.3 | Refseq |
| Affymetrix Human Transcriptome Array 2.0 | TC2100006967.hg.1 | NM_005534.3 | Refseq |
| Affymetrix Human Transcriptome Array 2.0 | TC2100006967.hg.1 | NM_005534.3 | Refseq |
| Affymetrix Human Transcriptome Array 2.0 | TC2100006967.hg.1 | NM_005534.3 | Refseq |
| Affymetrix Human Transcriptome Array 2.0 | TC2100006967.hg.1 | NM_005534.3 | Refseq |
| Affymetrix Human Transcriptome Array 2.0 | TC2100006967.hg.1 | NM_005534.3 | Refseq |
| Affymetrix Human Transcriptome Array 2.0 | TC2100006967.hg.1 | NM_005534.3 | Refseq |
| Affymetrix Human Transcriptome Array 2.0 | TC2100006967.hg.1 | NM_005534.3 | Refseq |
| Affymetrix Human Transcriptome Array 2.0 | TC2100006967.hg.1 | NM_005534.3 | Refseq |
| Affymetrix Human Transcriptome Array 2.0 | PSR21000942.hg.1 | ENST00000290219.6 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | PSR21000942.hg.1 | ENST00000381995.1 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | PSR21000942.hg.1 | ENST00000405436.1 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | PSR21000942.hg.1 | NM_005534.3 | Refseq |
| Affymetrix Human Transcriptome Array 2.0 | PSR21000942.hg.1 | TC21000129.hg.1 | HTA TC |
| Affymetrix Human Clariom D Assay | TC2100006967.hg.1 | ENST00000290219.10 | Ensembl |
| Affymetrix Human Clariom D Assay | TC2100006967.hg.1 | ENST00000290219.10 | Ensembl |
| Affymetrix Human Clariom D Assay | TC2100006967.hg.1 | ENST00000290219.10 | Ensembl |
| Affymetrix Human Clariom D Assay | TC2100006967.hg.1 | ENST00000290219.10 | Ensembl |
| Affymetrix Human Clariom D Assay | TC2100006967.hg.1 | ENST00000290219.10 | Ensembl |
| Affymetrix Human Clariom D Assay | TC2100006967.hg.1 | ENST00000290219.10 | Ensembl |
| Affymetrix Human Clariom D Assay | TC2100006967.hg.1 | ENST00000290219.10 | Ensembl |
| Affymetrix Human Clariom D Assay | TC2100006967.hg.1 | ENST00000290219.10 | Ensembl |
| Affymetrix Human Clariom D Assay | TC2100006967.hg.1 | ENST00000290219.10 | Ensembl |
| Affymetrix Human Clariom D Assay | TC2100006967.hg.1 | ENST00000290219.10 | Ensembl |
| Affymetrix Human Clariom D Assay | TC2100006967.hg.1 | ENST00000290219.10 | Ensembl |
| Affymetrix Human Clariom D Assay | TC2100006967.hg.1 | ENST00000290219.10 | Ensembl |
| Affymetrix Human Clariom D Assay | TC2100006967.hg.1 | ENST00000381995.5 | Ensembl |
| Affymetrix Human Clariom D Assay | TC2100006967.hg.1 | ENST00000381995.5 | Ensembl |
| Affymetrix Human Clariom D Assay | TC2100006967.hg.1 | ENST00000381995.5 | Ensembl |
| Affymetrix Human Clariom D Assay | TC2100006967.hg.1 | ENST00000381995.5 | Ensembl |
| Affymetrix Human Clariom D Assay | TC2100006967.hg.1 | ENST00000381995.5 | Ensembl |
| Affymetrix Human Clariom D Assay | TC2100006967.hg.1 | ENST00000381995.5 | Ensembl |
| Affymetrix Human Clariom D Assay | TC2100006967.hg.1 | ENST00000381995.5 | Ensembl |
| Affymetrix Human Clariom D Assay | TC2100006967.hg.1 | ENST00000381995.5 | Ensembl |
| Affymetrix Human Clariom D Assay | TC2100006967.hg.1 | ENST00000381995.5 | Ensembl |
| Affymetrix Human Clariom D Assay | TC2100006967.hg.1 | ENST00000381995.5 | Ensembl |
| Affymetrix Human Clariom D Assay | TC2100006967.hg.1 | ENST00000381995.5 | Ensembl |
| Affymetrix Human Clariom D Assay | TC2100006967.hg.1 | ENST00000381995.5 | Ensembl |
| Affymetrix Human Clariom D Assay | TC2100006967.hg.1 | ENST00000405436.5 | Ensembl |
| Affymetrix Human Clariom D Assay | TC2100006967.hg.1 | ENST00000405436.5 | Ensembl |
| Affymetrix Human Clariom D Assay | TC2100006967.hg.1 | ENST00000405436.5 | Ensembl |
| Affymetrix Human Clariom D Assay | TC2100006967.hg.1 | ENST00000405436.5 | Ensembl |
| Affymetrix Human Clariom D Assay | TC2100006967.hg.1 | ENST00000405436.5 | Ensembl |
| Affymetrix Human Clariom D Assay | TC2100006967.hg.1 | ENST00000405436.5 | Ensembl |
| Affymetrix Human Clariom D Assay | TC2100006967.hg.1 | ENST00000405436.5 | Ensembl |
| Affymetrix Human Clariom D Assay | TC2100006967.hg.1 | ENST00000405436.5 | Ensembl |
| Affymetrix Human Clariom D Assay | TC2100006967.hg.1 | ENST00000405436.5 | Ensembl |
| Affymetrix Human Clariom D Assay | TC2100006967.hg.1 | ENST00000405436.5 | Ensembl |
| Affymetrix Human Clariom D Assay | TC2100006967.hg.1 | ENST00000405436.5 | Ensembl |
| Affymetrix Human Clariom D Assay | TC2100006967.hg.1 | ENST00000421802.1 | Ensembl |
| Affymetrix Human Clariom D Assay | TC2100006967.hg.1 | ENST00000421802.1 | Ensembl |
| Affymetrix Human Clariom D Assay | TC2100006967.hg.1 | ENST00000439213.5 | Ensembl |
| Affymetrix Human Clariom D Assay | TC2100006967.hg.1 | ENST00000439213.5 | Ensembl |
| Affymetrix Human Clariom D Assay | TC2100006967.hg.1 | ENST00000439213.5 | Ensembl |
| Affymetrix Human Clariom D Assay | TC2100006967.hg.1 | ENST00000439213.5 | Ensembl |
| Affymetrix Human Clariom D Assay | TC2100006967.hg.1 | ENST00000439213.5 | Ensembl |
| Affymetrix Human Clariom D Assay | TC2100006967.hg.1 | ENST00000439213.5 | Ensembl |
| Affymetrix Human Clariom D Assay | TC2100006967.hg.1 | ENST00000439213.5 | Ensembl |
| Affymetrix Human Clariom D Assay | TC2100006967.hg.1 | ENST00000439213.5 | Ensembl |
| Affymetrix Human Clariom D Assay | TC2100006967.hg.1 | ENST00000439213.5 | Ensembl |
| Affymetrix Human Clariom D Assay | TC2100006967.hg.1 | ENST00000545369.2 | Ensembl |
| Affymetrix Human Clariom D Assay | TC2100006967.hg.1 | ENST00000545369.2 | Ensembl |
| Affymetrix Human Clariom D Assay | TC2100006967.hg.1 | ENST00000545369.2 | Ensembl |
| Affymetrix Human Clariom D Assay | TC2100006967.hg.1 | ENST00000545369.2 | Ensembl |
| Affymetrix Human Clariom D Assay | TC2100006967.hg.1 | ENST00000545369.2 | Ensembl |
| Affymetrix Human Clariom D Assay | TC2100006967.hg.1 | ENST00000545369.2 | Ensembl |
| Affymetrix Human Clariom D Assay | TC2100006967.hg.1 | ENST00000545369.2 | Ensembl |
| Affymetrix Human Clariom D Assay | TC2100006967.hg.1 | ENST00000545369.2 | Ensembl |
| Affymetrix Human Clariom D Assay | TC2100006967.hg.1 | ENST00000545369.2 | Ensembl |
| Affymetrix Human Clariom D Assay | TC2100006967.hg.1 | NM_005534.3 | Refseq |
| Affymetrix Human Clariom D Assay | TC2100006967.hg.1 | NM_005534.3 | Refseq |
| Affymetrix Human Clariom D Assay | TC2100006967.hg.1 | NM_005534.3 | Refseq |
| Affymetrix Human Clariom D Assay | TC2100006967.hg.1 | NM_005534.3 | Refseq |
| Affymetrix Human Clariom D Assay | TC2100006967.hg.1 | NM_005534.3 | Refseq |
| Affymetrix Human Clariom D Assay | TC2100006967.hg.1 | NM_005534.3 | Refseq |
| Affymetrix Human Clariom D Assay | TC2100006967.hg.1 | NM_005534.3 | Refseq |
| Affymetrix Human Clariom D Assay | TC2100006967.hg.1 | NM_005534.3 | Refseq |
| Affymetrix Human Clariom D Assay | TC2100006967.hg.1 | NM_005534.3 | Refseq |
| Affymetrix Human Clariom D Assay | TC2100006967.hg.1 | NM_005534.3 | Refseq |
| Affymetrix Human Clariom D Assay | TC2100006967.hg.1 | NM_005534.3 | Refseq |
| Affymetrix Human Clariom D Assay | TC2100006967.hg.1 | NM_005534.3 | Refseq |
| Affymetrix Human Clariom D Assay | PSR2100146781.hg.1 | ENST00000290219.10 | Ensembl |
| Affymetrix Human Clariom D Assay | PSR2100146781.hg.1 | ENST00000381995.5 | Ensembl |
| Affymetrix Human Clariom D Assay | PSR2100146781.hg.1 | ENST00000405436.5 | Ensembl |
| Affymetrix Human Clariom D Assay | PSR2100146781.hg.1 | ENST00000421802.1 | Ensembl |
| Affymetrix Human Clariom D Assay | PSR2100146781.hg.1 | ENST00000545369.2 | Ensembl |
| Affymetrix Human Clariom D Assay | PSR2100146781.hg.1 | NM_005534.3 | Refseq |
| Affymetrix Human Clariom D Assay | PSR2100146781.hg.1 | TC2100006967.hg.1 | Clariom D TC |
| Affymetrix Human Clariom D Assay | PSR2100146781.hg.1 | ENST00000290219.10 | Ensembl |
| Affymetrix Human Clariom D Assay | PSR2100146781.hg.1 | ENST00000381995.5 | Ensembl |
| Affymetrix Human Clariom D Assay | PSR2100146781.hg.1 | ENST00000405436.5 | Ensembl |
| Affymetrix Human Clariom D Assay | PSR2100146781.hg.1 | ENST00000421802.1 | Ensembl |
| Affymetrix Human Clariom D Assay | PSR2100146781.hg.1 | ENST00000545369.2 | Ensembl |
| Affymetrix Human Clariom D Assay | PSR2100146781.hg.1 | NM_005534.3 | Refseq |

IFNGR2
Genomic Map