Assay Details
| Gene Symbol: DUOXA1 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | NM_001276267.1 | NP_001263196.1 | 4 - 5 | 361 | 83 | |
| XM_011522183.2 | XP_011520485.1 | 4 - 5 | 414 | 83 | ||
| GenBank mRNA | BC020841.1 | - | 4 - 5 | 361 | 4689593 | 83 |
More Information
Additional Information:
This assay is in the 5'UTR region of the BC020841.1, NM_001276267.1, XM_011522183.2 transcripts.
Set Membership: |
|

DUOXA1
Genomic Map