| Array Name |
Probe IDs |
Targets |
Target Type |
| Affymetrix Human Transcriptome Array 2.0 |
TC0900010769.hg.1 |
ENST00000297689.3 |
Ensembl |
| Affymetrix Human Transcriptome Array 2.0 |
TC0900010769.hg.1 |
ENST00000297689.3 |
Ensembl |
| Affymetrix Human Transcriptome Array 2.0 |
TC0900010769.hg.1 |
ENST00000297689.3 |
Ensembl |
| Affymetrix Human Transcriptome Array 2.0 |
TC0900010769.hg.1 |
ENST00000297689.3 |
Ensembl |
| Affymetrix Human Transcriptome Array 2.0 |
TC0900010769.hg.1 |
ENST00000297689.3 |
Ensembl |
| Affymetrix Human Transcriptome Array 2.0 |
TC0900010769.hg.1 |
NM_001289999.1 |
Refseq |
| Affymetrix Human Transcriptome Array 2.0 |
TC0900010769.hg.1 |
NM_001289999.1 |
Refseq |
| Affymetrix Human Transcriptome Array 2.0 |
TC0900010769.hg.1 |
NM_001289999.1 |
Refseq |
| Affymetrix Human Transcriptome Array 2.0 |
TC0900010769.hg.1 |
NM_001289999.1 |
Refseq |
| Affymetrix Human Transcriptome Array 2.0 |
TC0900010769.hg.1 |
NM_001289999.1 |
Refseq |
| Affymetrix Human Transcriptome Array 2.0 |
TC0900010769.hg.1 |
NM_001289999.1 |
Refseq |
| Affymetrix Human Transcriptome Array 2.0 |
TC0900010769.hg.1 |
NM_001290000.1 |
Refseq |
| Affymetrix Human Transcriptome Array 2.0 |
TC0900010769.hg.1 |
NM_001290000.1 |
Refseq |
| Affymetrix Human Transcriptome Array 2.0 |
TC0900010769.hg.1 |
NM_001290000.1 |
Refseq |
| Affymetrix Human Transcriptome Array 2.0 |
TC0900010769.hg.1 |
NM_001290000.1 |
Refseq |
| Affymetrix Human Transcriptome Array 2.0 |
TC0900010769.hg.1 |
NM_001290000.1 |
Refseq |
| Affymetrix Human Transcriptome Array 2.0 |
TC0900010769.hg.1 |
NM_005384.2 |
Refseq |
| Affymetrix Human Transcriptome Array 2.0 |
TC0900010769.hg.1 |
NM_005384.2 |
Refseq |
| Affymetrix Human Transcriptome Array 2.0 |
TC0900010769.hg.1 |
NM_005384.2 |
Refseq |
| Affymetrix Human Transcriptome Array 2.0 |
TC0900010769.hg.1 |
NM_005384.2 |
Refseq |
| Affymetrix Human Transcriptome Array 2.0 |
TC0900010769.hg.1 |
NM_005384.2 |
Refseq |
| Affymetrix Human Transcriptome Array 2.0 |
PSR09016492.hg.1 |
TC09001325.hg.1 |
HTA TC |
| Affymetrix Human Transcriptome Array 2.0 |
PSR09016492.hg.1 |
ENST00000297689.2 |
Ensembl |
| Affymetrix Human Transcriptome Array 2.0 |
PSR09016492.hg.1 |
ENST00000375724.1 |
Ensembl |
| Affymetrix Human Transcriptome Array 2.0 |
PSR09016492.hg.1 |
NM_005384.2 |
Refseq |
| Affymetrix Human Transcriptome Array 2.0 |
PSR09016494.hg.1 |
TC09001325.hg.1 |
HTA TC |
| Affymetrix Human Transcriptome Array 2.0 |
PSR09016494.hg.1 |
ENST00000297689.2 |
Ensembl |
| Affymetrix Human Transcriptome Array 2.0 |
PSR09016494.hg.1 |
ENST00000375724.1 |
Ensembl |
| Affymetrix Human Transcriptome Array 2.0 |
PSR09016494.hg.1 |
NM_005384.2 |
Refseq |
| Affymetrix Human Clariom D Assay |
TC0900010769.hg.1 |
ENST00000297689.3 |
Ensembl |
| Affymetrix Human Clariom D Assay |
TC0900010769.hg.1 |
ENST00000297689.3 |
Ensembl |
| Affymetrix Human Clariom D Assay |
TC0900010769.hg.1 |
ENST00000297689.3 |
Ensembl |
| Affymetrix Human Clariom D Assay |
TC0900010769.hg.1 |
ENST00000297689.3 |
Ensembl |
| Affymetrix Human Clariom D Assay |
TC0900010769.hg.1 |
ENST00000297689.3 |
Ensembl |
| Affymetrix Human Clariom D Assay |
TC0900010769.hg.1 |
NM_001289999.1 |
Refseq |
| Affymetrix Human Clariom D Assay |
TC0900010769.hg.1 |
NM_001289999.1 |
Refseq |
| Affymetrix Human Clariom D Assay |
TC0900010769.hg.1 |
NM_001289999.1 |
Refseq |
| Affymetrix Human Clariom D Assay |
TC0900010769.hg.1 |
NM_001289999.1 |
Refseq |
| Affymetrix Human Clariom D Assay |
TC0900010769.hg.1 |
NM_001289999.1 |
Refseq |
| Affymetrix Human Clariom D Assay |
TC0900010769.hg.1 |
NM_001289999.1 |
Refseq |
| Affymetrix Human Clariom D Assay |
TC0900010769.hg.1 |
NM_001290000.1 |
Refseq |
| Affymetrix Human Clariom D Assay |
TC0900010769.hg.1 |
NM_001290000.1 |
Refseq |
| Affymetrix Human Clariom D Assay |
TC0900010769.hg.1 |
NM_001290000.1 |
Refseq |
| Affymetrix Human Clariom D Assay |
TC0900010769.hg.1 |
NM_001290000.1 |
Refseq |
| Affymetrix Human Clariom D Assay |
TC0900010769.hg.1 |
NM_001290000.1 |
Refseq |
| Affymetrix Human Clariom D Assay |
TC0900010769.hg.1 |
NM_005384.2 |
Refseq |
| Affymetrix Human Clariom D Assay |
TC0900010769.hg.1 |
NM_005384.2 |
Refseq |
| Affymetrix Human Clariom D Assay |
TC0900010769.hg.1 |
NM_005384.2 |
Refseq |
| Affymetrix Human Clariom D Assay |
TC0900010769.hg.1 |
NM_005384.2 |
Refseq |
| Affymetrix Human Clariom D Assay |
TC0900010769.hg.1 |
NM_005384.2 |
Refseq |
| Affymetrix Human Clariom D Assay |
PSR0900173219.hg.1 |
ENST00000297689.3 |
Ensembl |
| Affymetrix Human Clariom D Assay |
PSR0900173219.hg.1 |
NM_005384.2 |
Refseq |
| Affymetrix Human Clariom D Assay |
PSR0900173219.hg.1 |
ENST00000297689.3 |
Ensembl |
| Affymetrix Human Clariom D Assay |
PSR0900173219.hg.1 |
NM_005384.2 |
Refseq |
| Affymetrix Human Clariom D Assay |
PSR0900173219.hg.1 |
TC0900010769.hg.1 |
Clariom D TC |
| Affymetrix Human Clariom D Assay |
PSR0900173218.hg.1 |
ENST00000297689.3 |
Ensembl |
| Affymetrix Human Clariom D Assay |
PSR0900173218.hg.1 |
NM_001289999.1 |
Refseq |
| Affymetrix Human Clariom D Assay |
PSR0900173218.hg.1 |
NM_001290000.1 |
Refseq |
| Affymetrix Human Clariom D Assay |
PSR0900173218.hg.1 |
NM_005384.2 |
Refseq |
| Affymetrix Human Clariom D Assay |
PSR0900173218.hg.1 |
ENST00000297689.3 |
Ensembl |
| Affymetrix Human Clariom D Assay |
PSR0900173218.hg.1 |
NM_001289999.1 |
Refseq |
| Affymetrix Human Clariom D Assay |
PSR0900173218.hg.1 |
NM_001290000.1 |
Refseq |
| Affymetrix Human Clariom D Assay |
PSR0900173218.hg.1 |
NM_005384.2 |
Refseq |
| Affymetrix Human Clariom D Assay |
PSR0900173218.hg.1 |
TC0900010769.hg.1 |
Clariom D TC |