Assay Details
| Gene Symbol: BTAF1 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | NM_003972.2 | NP_003963.1 | 29 - 30 | 4267 | 139 | |
| XM_011540326.1 | XP_011538628.1 | 29 - 30 | 4366 | 139 | ||
| XM_011540327.2 | XP_011538629.1 | 26 - 27 | 4032 | 139 | ||
| XM_011540328.2 | XP_011538630.1 | 26 - 27 | 3860 | 139 | ||
| XM_017016877.1 | XP_016872366.1 | 28 - 29 | 4329 | 139 | ||
| XM_017016878.1 | XP_016872367.1 | 28 - 29 | 4159 | 139 | ||
| XM_017016879.1 | XP_016872368.1 | 28 - 29 | 4283 | 139 | ||
| XM_017016880.1 | XP_016872369.1 | 28 - 29 | 4287 | 139 | ||
| XM_017016881.1 | XP_016872370.1 | 27 - 28 | 4136 | 139 | ||
| GenBank mRNA | AB208843.1 | - | 17 - 18 | 2661 | 139 | |
| AF038362.1 | - | 29 - 30 | 4286 | 139 | ||
| AJ001017.2 | - | 29 - 30 | 4181 | 139 | ||
| AK303554.1 | - | 7 - 8 | 944 | 139 | ||
| BC029930.1 | - | 5 - 6 | 662 | 139 | ||
| BC112201.1 | - | 29 - 30 | 4251 | 8327669 | 139 | |
More Information
Updated Mapping Information
This assay targets the same location as HTA transcript cluster(s) TC10000655.hg.1
Set Membership: |
|

BTAF1
Genomic Map