Assay Details
| Gene Symbol: CAPN15 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | XM_011522620.2 | XP_011520922.1 | 4 | 1759 | 128 | |
| XM_011522621.2 | XP_011520923.1 | 4 | 2687 | 128 | ||
| XM_011522622.1 | XP_011520924.1 | 2 | 1391 | 128 | ||
| XM_011522626.2 | XP_011520928.1 | 4 | 2251 | 128 | ||
| XM_011522627.2 | XP_011520929.1 | 4 | 2256 | 128 | ||
| GenBank mRNA | BC032648.1 | - | 4 | 1612 | 128 | |
More Information
Additional Information:
This assay is in the 5'UTR region of the XM_011522620.2, XM_011522621.2, XM_011522622.1, XM_011522626.2, XM_011522627.2 transcripts.
Set Membership: |
|
Panther Classification:
Gene Ontology Categories:
MicroArray Probes:
Affymetrix
| Array Name | Probe IDs | Targets | Target Type |
|---|---|---|---|
| Affymetrix Human Transcriptome Array 2.0 | TC1600006469.hg.1 | ENST00000219611.6 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC1600006469.hg.1 | ENST00000219611.6 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC1600006469.hg.1 | ENST00000219611.6 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC1600006469.hg.1 | ENST00000219611.6 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC1600006469.hg.1 | ENST00000219611.6 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC1600006469.hg.1 | ENST00000219611.6 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC1600006469.hg.1 | ENST00000219611.6 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC1600006469.hg.1 | ENST00000219611.6 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC1600006469.hg.1 | ENST00000219611.6 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC1600006469.hg.1 | ENST00000219611.6 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC1600006469.hg.1 | ENST00000219611.6 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC1600006469.hg.1 | ENST00000219611.6 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC1600006469.hg.1 | ENST00000219611.6 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC1600006469.hg.1 | ENST00000219611.6 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC1600006469.hg.1 | ENST00000219611.6 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC1600006469.hg.1 | ENST00000219611.6 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC1600006469.hg.1 | ENST00000219611.6 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC1600006469.hg.1 | ENST00000219611.6 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC1600006469.hg.1 | ENST00000219611.6 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC1600006469.hg.1 | ENST00000562370.5 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC1600006469.hg.1 | ENST00000562370.5 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC1600006469.hg.1 | ENST00000562370.5 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC1600006469.hg.1 | ENST00000562370.5 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC1600006469.hg.1 | ENST00000562370.5 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC1600006469.hg.1 | ENST00000562370.5 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC1600006469.hg.1 | ENST00000565010.1 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC1600006469.hg.1 | ENST00000565010.1 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC1600006469.hg.1 | ENST00000565010.1 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC1600006469.hg.1 | ENST00000565010.1 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC1600006469.hg.1 | ENST00000565010.1 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC1600006469.hg.1 | ENST00000565010.1 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC1600006469.hg.1 | ENST00000565010.1 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC1600006469.hg.1 | ENST00000566977.1 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC1600006469.hg.1 | ENST00000566977.1 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC1600006469.hg.1 | ENST00000567216.5 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC1600006469.hg.1 | ENST00000567216.5 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC1600006469.hg.1 | ENST00000567216.5 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC1600006469.hg.1 | ENST00000567216.5 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC1600006469.hg.1 | ENST00000567216.5 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC1600006469.hg.1 | ENST00000567216.5 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC1600006469.hg.1 | ENST00000567216.5 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC1600006469.hg.1 | ENST00000568402.1 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC1600006469.hg.1 | ENST00000568402.1 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC1600006469.hg.1 | ENST00000568988.5 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC1600006469.hg.1 | ENST00000568988.5 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC1600006469.hg.1 | ENST00000568988.5 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC1600006469.hg.1 | ENST00000568988.5 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC1600006469.hg.1 | ENST00000581803.1 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC1600006469.hg.1 | NM_005632.2 | Refseq |
| Affymetrix Human Transcriptome Array 2.0 | TC1600006469.hg.1 | NM_005632.2 | Refseq |
| Affymetrix Human Transcriptome Array 2.0 | TC1600006469.hg.1 | NM_005632.2 | Refseq |
| Affymetrix Human Transcriptome Array 2.0 | TC1600006469.hg.1 | NM_005632.2 | Refseq |
| Affymetrix Human Transcriptome Array 2.0 | TC1600006469.hg.1 | NM_005632.2 | Refseq |
| Affymetrix Human Transcriptome Array 2.0 | TC1600006469.hg.1 | NM_005632.2 | Refseq |
| Affymetrix Human Transcriptome Array 2.0 | TC1600006469.hg.1 | NM_005632.2 | Refseq |
| Affymetrix Human Transcriptome Array 2.0 | TC1600006469.hg.1 | NM_005632.2 | Refseq |
| Affymetrix Human Transcriptome Array 2.0 | TC1600006469.hg.1 | NM_005632.2 | Refseq |
| Affymetrix Human Transcriptome Array 2.0 | TC1600006469.hg.1 | NM_005632.2 | Refseq |
| Affymetrix Human Transcriptome Array 2.0 | TC1600006469.hg.1 | NM_005632.2 | Refseq |
| Affymetrix Human Transcriptome Array 2.0 | TC1600006469.hg.1 | NM_005632.2 | Refseq |
| Affymetrix Human Transcriptome Array 2.0 | TC1600006469.hg.1 | NM_005632.2 | Refseq |
| Affymetrix Human Transcriptome Array 2.0 | TC1600006469.hg.1 | NM_005632.2 | Refseq |
| Affymetrix Human Transcriptome Array 2.0 | TC1600006469.hg.1 | NM_005632.2 | Refseq |
| Affymetrix Human Transcriptome Array 2.0 | TC1600006469.hg.1 | NM_005632.2 | Refseq |
| Affymetrix Human Transcriptome Array 2.0 | TC1600006469.hg.1 | NM_005632.2 | Refseq |
| Affymetrix Human Transcriptome Array 2.0 | TC1600006469.hg.1 | NM_005632.2 | Refseq |
| Affymetrix Human Transcriptome Array 2.0 | TC1600006469.hg.1 | NM_005632.2 | Refseq |
| Affymetrix Human Transcriptome Array 2.0 | TC1600006469.hg.1 | NR_049852.1 | Refseq |
| Affymetrix Human Clariom D Assay | TC1600006469.hg.1 | ENST00000219611.6 | Ensembl |
| Affymetrix Human Clariom D Assay | TC1600006469.hg.1 | ENST00000219611.6 | Ensembl |
| Affymetrix Human Clariom D Assay | TC1600006469.hg.1 | ENST00000219611.6 | Ensembl |
| Affymetrix Human Clariom D Assay | TC1600006469.hg.1 | ENST00000219611.6 | Ensembl |
| Affymetrix Human Clariom D Assay | TC1600006469.hg.1 | ENST00000219611.6 | Ensembl |
| Affymetrix Human Clariom D Assay | TC1600006469.hg.1 | ENST00000219611.6 | Ensembl |
| Affymetrix Human Clariom D Assay | TC1600006469.hg.1 | ENST00000219611.6 | Ensembl |
| Affymetrix Human Clariom D Assay | TC1600006469.hg.1 | ENST00000219611.6 | Ensembl |
| Affymetrix Human Clariom D Assay | TC1600006469.hg.1 | ENST00000219611.6 | Ensembl |
| Affymetrix Human Clariom D Assay | TC1600006469.hg.1 | ENST00000219611.6 | Ensembl |
| Affymetrix Human Clariom D Assay | TC1600006469.hg.1 | ENST00000219611.6 | Ensembl |
| Affymetrix Human Clariom D Assay | TC1600006469.hg.1 | ENST00000219611.6 | Ensembl |
| Affymetrix Human Clariom D Assay | TC1600006469.hg.1 | ENST00000219611.6 | Ensembl |
| Affymetrix Human Clariom D Assay | TC1600006469.hg.1 | ENST00000219611.6 | Ensembl |
| Affymetrix Human Clariom D Assay | TC1600006469.hg.1 | ENST00000219611.6 | Ensembl |
| Affymetrix Human Clariom D Assay | TC1600006469.hg.1 | ENST00000219611.6 | Ensembl |
| Affymetrix Human Clariom D Assay | TC1600006469.hg.1 | ENST00000219611.6 | Ensembl |
| Affymetrix Human Clariom D Assay | TC1600006469.hg.1 | ENST00000219611.6 | Ensembl |
| Affymetrix Human Clariom D Assay | TC1600006469.hg.1 | ENST00000219611.6 | Ensembl |
| Affymetrix Human Clariom D Assay | TC1600006469.hg.1 | ENST00000562370.5 | Ensembl |
| Affymetrix Human Clariom D Assay | TC1600006469.hg.1 | ENST00000562370.5 | Ensembl |
| Affymetrix Human Clariom D Assay | TC1600006469.hg.1 | ENST00000562370.5 | Ensembl |
| Affymetrix Human Clariom D Assay | TC1600006469.hg.1 | ENST00000562370.5 | Ensembl |
| Affymetrix Human Clariom D Assay | TC1600006469.hg.1 | ENST00000562370.5 | Ensembl |
| Affymetrix Human Clariom D Assay | TC1600006469.hg.1 | ENST00000562370.5 | Ensembl |
| Affymetrix Human Clariom D Assay | TC1600006469.hg.1 | ENST00000565010.1 | Ensembl |
| Affymetrix Human Clariom D Assay | TC1600006469.hg.1 | ENST00000565010.1 | Ensembl |
| Affymetrix Human Clariom D Assay | TC1600006469.hg.1 | ENST00000565010.1 | Ensembl |
| Affymetrix Human Clariom D Assay | TC1600006469.hg.1 | ENST00000565010.1 | Ensembl |
| Affymetrix Human Clariom D Assay | TC1600006469.hg.1 | ENST00000565010.1 | Ensembl |
| Affymetrix Human Clariom D Assay | TC1600006469.hg.1 | ENST00000565010.1 | Ensembl |
| Affymetrix Human Clariom D Assay | TC1600006469.hg.1 | ENST00000565010.1 | Ensembl |
| Affymetrix Human Clariom D Assay | TC1600006469.hg.1 | ENST00000566977.1 | Ensembl |
| Affymetrix Human Clariom D Assay | TC1600006469.hg.1 | ENST00000566977.1 | Ensembl |
| Affymetrix Human Clariom D Assay | TC1600006469.hg.1 | ENST00000567216.5 | Ensembl |
| Affymetrix Human Clariom D Assay | TC1600006469.hg.1 | ENST00000567216.5 | Ensembl |
| Affymetrix Human Clariom D Assay | TC1600006469.hg.1 | ENST00000567216.5 | Ensembl |
| Affymetrix Human Clariom D Assay | TC1600006469.hg.1 | ENST00000567216.5 | Ensembl |
| Affymetrix Human Clariom D Assay | TC1600006469.hg.1 | ENST00000567216.5 | Ensembl |
| Affymetrix Human Clariom D Assay | TC1600006469.hg.1 | ENST00000567216.5 | Ensembl |
| Affymetrix Human Clariom D Assay | TC1600006469.hg.1 | ENST00000567216.5 | Ensembl |
| Affymetrix Human Clariom D Assay | TC1600006469.hg.1 | ENST00000568402.1 | Ensembl |
| Affymetrix Human Clariom D Assay | TC1600006469.hg.1 | ENST00000568402.1 | Ensembl |
| Affymetrix Human Clariom D Assay | TC1600006469.hg.1 | ENST00000568988.5 | Ensembl |
| Affymetrix Human Clariom D Assay | TC1600006469.hg.1 | ENST00000568988.5 | Ensembl |
| Affymetrix Human Clariom D Assay | TC1600006469.hg.1 | ENST00000568988.5 | Ensembl |
| Affymetrix Human Clariom D Assay | TC1600006469.hg.1 | ENST00000568988.5 | Ensembl |
| Affymetrix Human Clariom D Assay | TC1600006469.hg.1 | ENST00000581803.1 | Ensembl |
| Affymetrix Human Clariom D Assay | TC1600006469.hg.1 | NM_005632.2 | Refseq |
| Affymetrix Human Clariom D Assay | TC1600006469.hg.1 | NM_005632.2 | Refseq |
| Affymetrix Human Clariom D Assay | TC1600006469.hg.1 | NM_005632.2 | Refseq |
| Affymetrix Human Clariom D Assay | TC1600006469.hg.1 | NM_005632.2 | Refseq |
| Affymetrix Human Clariom D Assay | TC1600006469.hg.1 | NM_005632.2 | Refseq |
| Affymetrix Human Clariom D Assay | TC1600006469.hg.1 | NM_005632.2 | Refseq |
| Affymetrix Human Clariom D Assay | TC1600006469.hg.1 | NM_005632.2 | Refseq |
| Affymetrix Human Clariom D Assay | TC1600006469.hg.1 | NM_005632.2 | Refseq |
| Affymetrix Human Clariom D Assay | TC1600006469.hg.1 | NM_005632.2 | Refseq |
| Affymetrix Human Clariom D Assay | TC1600006469.hg.1 | NM_005632.2 | Refseq |
| Affymetrix Human Clariom D Assay | TC1600006469.hg.1 | NM_005632.2 | Refseq |
| Affymetrix Human Clariom D Assay | TC1600006469.hg.1 | NM_005632.2 | Refseq |
| Affymetrix Human Clariom D Assay | TC1600006469.hg.1 | NM_005632.2 | Refseq |
| Affymetrix Human Clariom D Assay | TC1600006469.hg.1 | NM_005632.2 | Refseq |
| Affymetrix Human Clariom D Assay | TC1600006469.hg.1 | NM_005632.2 | Refseq |
| Affymetrix Human Clariom D Assay | TC1600006469.hg.1 | NM_005632.2 | Refseq |
| Affymetrix Human Clariom D Assay | TC1600006469.hg.1 | NM_005632.2 | Refseq |
| Affymetrix Human Clariom D Assay | TC1600006469.hg.1 | NM_005632.2 | Refseq |
| Affymetrix Human Clariom D Assay | TC1600006469.hg.1 | NM_005632.2 | Refseq |
| Affymetrix Human Clariom D Assay | TC1600006469.hg.1 | NR_049852.1 | Refseq |

CAPN15
Genomic Map