Assay Details
| Gene Symbol: SCUBE1 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | NM_173050.3 | NP_766638.2 | 13 - 14 | 1672 | 71 | |
| XM_005261752.1 | XP_005261809.1 | 14 - 15 | 1723 | 71 | ||
| XM_011530389.2 | XP_011528691.1 | 14 - 15 | 1717 | 71 | ||
| XM_011530390.2 | XP_011528692.1 | 13 - 14 | 1602 | 71 | ||
| GenBank mRNA | AF525689.1 | - | 14 - 15 | 1565 | 71 | |
| AK127719.1 | - | 10 - 11 | 1145 | 71 | ||
More Information
Updated Mapping Information
This assay targets the same location as HTA transcript cluster(s) TC22000861.hg.1
Set Membership: |
|
Matching Ambion siRNAs:
Panther Classification:
Gene Ontology Categories:
MicroArray Probes:
Affymetrix
| Array Name | Probe IDs | Targets | Target Type |
|---|---|---|---|
| Affymetrix Human Transcriptome Array 2.0 | PSR22014552.hg.1 | ENST00000360835.3 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | PSR22014552.hg.1 | ENST00000381243.3 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | PSR22014552.hg.1 | NM_173050.3 | Refseq |
| Affymetrix Human Transcriptome Array 2.0 | PSR22014552.hg.1 | TC22000861.hg.1 | HTA TC |
| Affymetrix Human Transcriptome Array 2.0 | PSR22014551.hg.1 | ENST00000360835.3 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | PSR22014551.hg.1 | ENST00000381243.3 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | PSR22014551.hg.1 | NM_173050.3 | Refseq |
| Affymetrix Human Transcriptome Array 2.0 | PSR22014551.hg.1 | TC22000861.hg.1 | HTA TC |
| Affymetrix Human Transcriptome Array 2.0 | TC2200008907.hg.1 | ENST00000290460.7 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC2200008907.hg.1 | ENST00000290460.7 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC2200008907.hg.1 | ENST00000290460.7 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC2200008907.hg.1 | ENST00000290460.7 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC2200008907.hg.1 | ENST00000290460.7 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC2200008907.hg.1 | ENST00000290460.7 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC2200008907.hg.1 | ENST00000290460.7 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC2200008907.hg.1 | ENST00000290460.7 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC2200008907.hg.1 | ENST00000290460.7 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC2200008907.hg.1 | ENST00000290460.7 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC2200008907.hg.1 | ENST00000290460.7 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC2200008907.hg.1 | ENST00000360835.8 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC2200008907.hg.1 | ENST00000360835.8 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC2200008907.hg.1 | ENST00000360835.8 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC2200008907.hg.1 | ENST00000360835.8 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC2200008907.hg.1 | ENST00000360835.8 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC2200008907.hg.1 | ENST00000360835.8 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC2200008907.hg.1 | ENST00000360835.8 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC2200008907.hg.1 | ENST00000360835.8 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC2200008907.hg.1 | ENST00000360835.8 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC2200008907.hg.1 | ENST00000360835.8 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC2200008907.hg.1 | ENST00000360835.8 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC2200008907.hg.1 | ENST00000360835.8 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC2200008907.hg.1 | ENST00000360835.8 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC2200008907.hg.1 | ENST00000360835.8 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC2200008907.hg.1 | ENST00000360835.8 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC2200008907.hg.1 | ENST00000360835.8 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC2200008907.hg.1 | ENST00000360835.8 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC2200008907.hg.1 | ENST00000360835.8 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC2200008907.hg.1 | ENST00000360835.8 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC2200008907.hg.1 | ENST00000360835.8 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC2200008907.hg.1 | ENST00000360835.8 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC2200008907.hg.1 | ENST00000360835.8 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC2200008907.hg.1 | ENST00000360835.8 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC2200008907.hg.1 | ENST00000360835.8 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC2200008907.hg.1 | ENST00000360835.8 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC2200008907.hg.1 | ENST00000360835.8 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC2200008907.hg.1 | ENST00000360835.8 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC2200008907.hg.1 | ENST00000360835.8 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC2200008907.hg.1 | ENST00000360835.8 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC2200008907.hg.1 | ENST00000360835.8 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC2200008907.hg.1 | ENST00000449304.5 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC2200008907.hg.1 | ENST00000449304.5 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC2200008907.hg.1 | ENST00000449304.5 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC2200008907.hg.1 | ENST00000449304.5 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC2200008907.hg.1 | ENST00000449304.5 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC2200008907.hg.1 | ENST00000461595.5 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC2200008907.hg.1 | ENST00000461595.5 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC2200008907.hg.1 | ENST00000461595.5 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC2200008907.hg.1 | ENST00000461595.5 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC2200008907.hg.1 | ENST00000480833.1 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC2200008907.hg.1 | ENST00000480833.1 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC2200008907.hg.1 | ENST00000615096.4 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC2200008907.hg.1 | ENST00000615096.4 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC2200008907.hg.1 | ENST00000615096.4 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC2200008907.hg.1 | ENST00000615096.4 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC2200008907.hg.1 | ENST00000615096.4 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC2200008907.hg.1 | ENST00000615096.4 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC2200008907.hg.1 | ENST00000615096.4 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC2200008907.hg.1 | ENST00000615096.4 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC2200008907.hg.1 | ENST00000615096.4 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC2200008907.hg.1 | ENST00000615096.4 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC2200008907.hg.1 | ENST00000615096.4 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC2200008907.hg.1 | NM_173050.3 | Refseq |
| Affymetrix Human Transcriptome Array 2.0 | TC2200008907.hg.1 | NM_173050.3 | Refseq |
| Affymetrix Human Transcriptome Array 2.0 | TC2200008907.hg.1 | NM_173050.3 | Refseq |
| Affymetrix Human Transcriptome Array 2.0 | TC2200008907.hg.1 | NM_173050.3 | Refseq |
| Affymetrix Human Transcriptome Array 2.0 | TC2200008907.hg.1 | NM_173050.3 | Refseq |
| Affymetrix Human Transcriptome Array 2.0 | TC2200008907.hg.1 | NM_173050.3 | Refseq |
| Affymetrix Human Transcriptome Array 2.0 | TC2200008907.hg.1 | NM_173050.3 | Refseq |
| Affymetrix Human Transcriptome Array 2.0 | TC2200008907.hg.1 | NM_173050.3 | Refseq |
| Affymetrix Human Transcriptome Array 2.0 | TC2200008907.hg.1 | NM_173050.3 | Refseq |
| Affymetrix Human Transcriptome Array 2.0 | TC2200008907.hg.1 | NM_173050.3 | Refseq |
| Affymetrix Human Transcriptome Array 2.0 | TC2200008907.hg.1 | NM_173050.3 | Refseq |
| Affymetrix Human Transcriptome Array 2.0 | TC2200008907.hg.1 | NM_173050.3 | Refseq |
| Affymetrix Human Transcriptome Array 2.0 | TC2200008907.hg.1 | NM_173050.3 | Refseq |
| Affymetrix Human Transcriptome Array 2.0 | TC2200008907.hg.1 | NM_173050.3 | Refseq |
| Affymetrix Human Transcriptome Array 2.0 | TC2200008907.hg.1 | NM_173050.3 | Refseq |
| Affymetrix Human Transcriptome Array 2.0 | TC2200008907.hg.1 | NM_173050.3 | Refseq |
| Affymetrix Human Transcriptome Array 2.0 | TC2200008907.hg.1 | NM_173050.3 | Refseq |
| Affymetrix Human Transcriptome Array 2.0 | TC2200008907.hg.1 | NM_173050.3 | Refseq |
| Affymetrix Human Transcriptome Array 2.0 | TC2200008907.hg.1 | NM_173050.3 | Refseq |
| Affymetrix Human Transcriptome Array 2.0 | TC2200008907.hg.1 | NM_173050.3 | Refseq |
| Affymetrix Human Transcriptome Array 2.0 | TC2200008907.hg.1 | NM_173050.3 | Refseq |
| Affymetrix Human Transcriptome Array 2.0 | TC2200008907.hg.1 | NM_173050.3 | Refseq |
| Affymetrix Human Transcriptome Array 2.0 | TC2200008907.hg.1 | NM_173050.3 | Refseq |
| Affymetrix Human Transcriptome Array 2.0 | TC2200008907.hg.1 | NM_173050.3 | Refseq |
| Affymetrix Human Transcriptome Array 2.0 | TC2200008907.hg.1 | NM_173050.3 | Refseq |
| Affymetrix Human Transcriptome Array 2.0 | TC2200008907.hg.1 | NM_173050.3 | Refseq |
| Affymetrix Human Transcriptome Array 2.0 | TC2200008907.hg.1 | NM_173050.3 | Refseq |
| Affymetrix Human Transcriptome Array 2.0 | TC2200008907.hg.1 | NM_173050.3 | Refseq |
| Affymetrix Human Clariom D Assay | PSR2200164644.hg.1 | TC2200008907.hg.1 | Clariom D TC |
| Affymetrix Human Clariom D Assay | PSR2200164644.hg.1 | ENST00000360835.8 | Ensembl |
| Affymetrix Human Clariom D Assay | PSR2200164644.hg.1 | NM_173050.3 | Refseq |
| Affymetrix Human Clariom D Assay | PSR2200164644.hg.1 | ENST00000360835.8 | Ensembl |
| Affymetrix Human Clariom D Assay | PSR2200164644.hg.1 | NM_173050.3 | Refseq |
| Affymetrix Human Clariom D Assay | TC2200008907.hg.1 | ENST00000290460.7 | Ensembl |
| Affymetrix Human Clariom D Assay | TC2200008907.hg.1 | ENST00000290460.7 | Ensembl |
| Affymetrix Human Clariom D Assay | TC2200008907.hg.1 | ENST00000290460.7 | Ensembl |
| Affymetrix Human Clariom D Assay | TC2200008907.hg.1 | ENST00000290460.7 | Ensembl |
| Affymetrix Human Clariom D Assay | TC2200008907.hg.1 | ENST00000290460.7 | Ensembl |
| Affymetrix Human Clariom D Assay | TC2200008907.hg.1 | ENST00000290460.7 | Ensembl |
| Affymetrix Human Clariom D Assay | TC2200008907.hg.1 | ENST00000290460.7 | Ensembl |
| Affymetrix Human Clariom D Assay | TC2200008907.hg.1 | ENST00000290460.7 | Ensembl |
| Affymetrix Human Clariom D Assay | TC2200008907.hg.1 | ENST00000290460.7 | Ensembl |
| Affymetrix Human Clariom D Assay | TC2200008907.hg.1 | ENST00000290460.7 | Ensembl |
| Affymetrix Human Clariom D Assay | TC2200008907.hg.1 | ENST00000290460.7 | Ensembl |
| Affymetrix Human Clariom D Assay | TC2200008907.hg.1 | ENST00000360835.8 | Ensembl |
| Affymetrix Human Clariom D Assay | TC2200008907.hg.1 | ENST00000360835.8 | Ensembl |
| Affymetrix Human Clariom D Assay | TC2200008907.hg.1 | ENST00000360835.8 | Ensembl |
| Affymetrix Human Clariom D Assay | TC2200008907.hg.1 | ENST00000360835.8 | Ensembl |
| Affymetrix Human Clariom D Assay | TC2200008907.hg.1 | ENST00000360835.8 | Ensembl |
| Affymetrix Human Clariom D Assay | TC2200008907.hg.1 | ENST00000360835.8 | Ensembl |
| Affymetrix Human Clariom D Assay | TC2200008907.hg.1 | ENST00000360835.8 | Ensembl |
| Affymetrix Human Clariom D Assay | TC2200008907.hg.1 | ENST00000360835.8 | Ensembl |
| Affymetrix Human Clariom D Assay | TC2200008907.hg.1 | ENST00000360835.8 | Ensembl |
| Affymetrix Human Clariom D Assay | TC2200008907.hg.1 | ENST00000360835.8 | Ensembl |
| Affymetrix Human Clariom D Assay | TC2200008907.hg.1 | ENST00000360835.8 | Ensembl |
| Affymetrix Human Clariom D Assay | TC2200008907.hg.1 | ENST00000360835.8 | Ensembl |
| Affymetrix Human Clariom D Assay | TC2200008907.hg.1 | ENST00000360835.8 | Ensembl |
| Affymetrix Human Clariom D Assay | TC2200008907.hg.1 | ENST00000360835.8 | Ensembl |
| Affymetrix Human Clariom D Assay | TC2200008907.hg.1 | ENST00000360835.8 | Ensembl |
| Affymetrix Human Clariom D Assay | TC2200008907.hg.1 | ENST00000360835.8 | Ensembl |
| Affymetrix Human Clariom D Assay | TC2200008907.hg.1 | ENST00000360835.8 | Ensembl |
| Affymetrix Human Clariom D Assay | TC2200008907.hg.1 | ENST00000360835.8 | Ensembl |
| Affymetrix Human Clariom D Assay | TC2200008907.hg.1 | ENST00000360835.8 | Ensembl |
| Affymetrix Human Clariom D Assay | TC2200008907.hg.1 | ENST00000360835.8 | Ensembl |
| Affymetrix Human Clariom D Assay | TC2200008907.hg.1 | ENST00000360835.8 | Ensembl |
| Affymetrix Human Clariom D Assay | TC2200008907.hg.1 | ENST00000360835.8 | Ensembl |
| Affymetrix Human Clariom D Assay | TC2200008907.hg.1 | ENST00000360835.8 | Ensembl |
| Affymetrix Human Clariom D Assay | TC2200008907.hg.1 | ENST00000360835.8 | Ensembl |
| Affymetrix Human Clariom D Assay | TC2200008907.hg.1 | ENST00000360835.8 | Ensembl |
| Affymetrix Human Clariom D Assay | TC2200008907.hg.1 | ENST00000360835.8 | Ensembl |
| Affymetrix Human Clariom D Assay | TC2200008907.hg.1 | ENST00000360835.8 | Ensembl |
| Affymetrix Human Clariom D Assay | TC2200008907.hg.1 | ENST00000360835.8 | Ensembl |
| Affymetrix Human Clariom D Assay | TC2200008907.hg.1 | ENST00000360835.8 | Ensembl |
| Affymetrix Human Clariom D Assay | TC2200008907.hg.1 | ENST00000360835.8 | Ensembl |
| Affymetrix Human Clariom D Assay | TC2200008907.hg.1 | ENST00000449304.5 | Ensembl |
| Affymetrix Human Clariom D Assay | TC2200008907.hg.1 | ENST00000449304.5 | Ensembl |
| Affymetrix Human Clariom D Assay | TC2200008907.hg.1 | ENST00000449304.5 | Ensembl |
| Affymetrix Human Clariom D Assay | TC2200008907.hg.1 | ENST00000449304.5 | Ensembl |
| Affymetrix Human Clariom D Assay | TC2200008907.hg.1 | ENST00000449304.5 | Ensembl |
| Affymetrix Human Clariom D Assay | TC2200008907.hg.1 | ENST00000461595.5 | Ensembl |
| Affymetrix Human Clariom D Assay | TC2200008907.hg.1 | ENST00000461595.5 | Ensembl |
| Affymetrix Human Clariom D Assay | TC2200008907.hg.1 | ENST00000461595.5 | Ensembl |
| Affymetrix Human Clariom D Assay | TC2200008907.hg.1 | ENST00000461595.5 | Ensembl |
| Affymetrix Human Clariom D Assay | TC2200008907.hg.1 | ENST00000480833.1 | Ensembl |
| Affymetrix Human Clariom D Assay | TC2200008907.hg.1 | ENST00000480833.1 | Ensembl |
| Affymetrix Human Clariom D Assay | TC2200008907.hg.1 | ENST00000615096.4 | Ensembl |
| Affymetrix Human Clariom D Assay | TC2200008907.hg.1 | ENST00000615096.4 | Ensembl |
| Affymetrix Human Clariom D Assay | TC2200008907.hg.1 | ENST00000615096.4 | Ensembl |
| Affymetrix Human Clariom D Assay | TC2200008907.hg.1 | ENST00000615096.4 | Ensembl |
| Affymetrix Human Clariom D Assay | TC2200008907.hg.1 | ENST00000615096.4 | Ensembl |
| Affymetrix Human Clariom D Assay | TC2200008907.hg.1 | ENST00000615096.4 | Ensembl |
| Affymetrix Human Clariom D Assay | TC2200008907.hg.1 | ENST00000615096.4 | Ensembl |
| Affymetrix Human Clariom D Assay | TC2200008907.hg.1 | ENST00000615096.4 | Ensembl |
| Affymetrix Human Clariom D Assay | TC2200008907.hg.1 | ENST00000615096.4 | Ensembl |
| Affymetrix Human Clariom D Assay | TC2200008907.hg.1 | ENST00000615096.4 | Ensembl |
| Affymetrix Human Clariom D Assay | TC2200008907.hg.1 | ENST00000615096.4 | Ensembl |
| Affymetrix Human Clariom D Assay | TC2200008907.hg.1 | NM_173050.3 | Refseq |
| Affymetrix Human Clariom D Assay | TC2200008907.hg.1 | NM_173050.3 | Refseq |
| Affymetrix Human Clariom D Assay | TC2200008907.hg.1 | NM_173050.3 | Refseq |
| Affymetrix Human Clariom D Assay | TC2200008907.hg.1 | NM_173050.3 | Refseq |
| Affymetrix Human Clariom D Assay | TC2200008907.hg.1 | NM_173050.3 | Refseq |
| Affymetrix Human Clariom D Assay | TC2200008907.hg.1 | NM_173050.3 | Refseq |
| Affymetrix Human Clariom D Assay | TC2200008907.hg.1 | NM_173050.3 | Refseq |
| Affymetrix Human Clariom D Assay | TC2200008907.hg.1 | NM_173050.3 | Refseq |
| Affymetrix Human Clariom D Assay | TC2200008907.hg.1 | NM_173050.3 | Refseq |
| Affymetrix Human Clariom D Assay | TC2200008907.hg.1 | NM_173050.3 | Refseq |
| Affymetrix Human Clariom D Assay | TC2200008907.hg.1 | NM_173050.3 | Refseq |
| Affymetrix Human Clariom D Assay | TC2200008907.hg.1 | NM_173050.3 | Refseq |
| Affymetrix Human Clariom D Assay | TC2200008907.hg.1 | NM_173050.3 | Refseq |
| Affymetrix Human Clariom D Assay | TC2200008907.hg.1 | NM_173050.3 | Refseq |
| Affymetrix Human Clariom D Assay | TC2200008907.hg.1 | NM_173050.3 | Refseq |
| Affymetrix Human Clariom D Assay | TC2200008907.hg.1 | NM_173050.3 | Refseq |
| Affymetrix Human Clariom D Assay | TC2200008907.hg.1 | NM_173050.3 | Refseq |
| Affymetrix Human Clariom D Assay | TC2200008907.hg.1 | NM_173050.3 | Refseq |
| Affymetrix Human Clariom D Assay | TC2200008907.hg.1 | NM_173050.3 | Refseq |
| Affymetrix Human Clariom D Assay | TC2200008907.hg.1 | NM_173050.3 | Refseq |
| Affymetrix Human Clariom D Assay | TC2200008907.hg.1 | NM_173050.3 | Refseq |
| Affymetrix Human Clariom D Assay | TC2200008907.hg.1 | NM_173050.3 | Refseq |
| Affymetrix Human Clariom D Assay | TC2200008907.hg.1 | NM_173050.3 | Refseq |
| Affymetrix Human Clariom D Assay | TC2200008907.hg.1 | NM_173050.3 | Refseq |
| Affymetrix Human Clariom D Assay | TC2200008907.hg.1 | NM_173050.3 | Refseq |
| Affymetrix Human Clariom D Assay | TC2200008907.hg.1 | NM_173050.3 | Refseq |
| Affymetrix Human Clariom D Assay | TC2200008907.hg.1 | NM_173050.3 | Refseq |
| Affymetrix Human Clariom D Assay | TC2200008907.hg.1 | NM_173050.3 | Refseq |

SCUBE1
Genomic Map