Assay Details
| Gene Symbol: HOMER1 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | NM_001277077.1 | NP_001264006.1 | 1 - 2 | 1064 | 81 | |
| NM_001277078.1 | NP_001264007.1 | 1 - 2 | 1064 | 81 | ||
| NM_004272.4 | NP_004263.1 | 1 - 2 | 1064 | 81 | ||
| XM_017010059.1 | XP_016865548.1 | 1 - 2 | 1796 | 81 | ||
| GenBank mRNA | AK223471.1 | - | 1 - 2 | 326 | 81 | |
| AK312481.1 | - | 1 - 2 | 329 | 81 | ||
| BC015502.1 | - | 1 - 2 | 279 | 3904242 | 81 | |
| BC047438.1 | - | 1 - 2 | 1064 | 5294849 | 81 | |
| BU933636.1 | - | 1 - 2 | 632 | 81 | ||
| Y17829.1 | - | 1 - 2 | 77 | 81 | ||
More Information
Updated Mapping Information
This assay targets the same location as HTA transcript cluster(s) TC05001522.hg.1
Additional Information:
This assay is in the 5'UTR region of the AK312481.1,BC015502.1,BC047438.1,NM_001277077.1,NM_001277078.1,NM_004272.4,XM_017010059.1,Y17829.1 transcripts.
Set Membership: |
|
Matching Ambion siRNAs:
Gene Ontology Categories:
Function(s)
Process(es)
skeletal muscle contraction
phospholipase C-activating G-protein coupled glutamate receptor signaling pathway
chemical synaptic transmission
positive regulation of signal transduction
behavioral response to cocaine
skeletal muscle fiber development
chemical homeostasis within a tissue
response to calcium ion
positive regulation of calcium ion transport
regulation of calcium ion import
regulation of store-operated calcium entry
regulation of cation channel activity
protein binding
G-protein coupled glutamate receptor binding
signaling adaptor activity
ion channel binding
skeletal muscle contraction
phospholipase C-activating G-protein coupled glutamate receptor signaling pathway
chemical synaptic transmission
positive regulation of signal transduction
behavioral response to cocaine
skeletal muscle fiber development
chemical homeostasis within a tissue
response to calcium ion
positive regulation of calcium ion transport
regulation of calcium ion import
regulation of store-operated calcium entry
regulation of cation channel activity
protein binding
G-protein coupled glutamate receptor binding
signaling adaptor activity
ion channel binding
MicroArray Probes:
Affymetrix
ABI
| Array Name | Probe IDs | Targets | Target Type |
|---|---|---|---|
| Affymetrix Human Transcriptome Array 2.0 | PSR05020877.hg.1 | TC05001522.hg.1 | HTA TC |
| Affymetrix Human Transcriptome Array 2.0 | PSR05020877.hg.1 | ENST00000334082.6 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | PSR05020877.hg.1 | NM_004272.4 | Refseq |
| Affymetrix Human Transcriptome Array 2.0 | TC0500011240.hg.1 | ENST00000282260.10 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0500011240.hg.1 | ENST00000282260.10 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0500011240.hg.1 | ENST00000282260.10 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0500011240.hg.1 | ENST00000282260.10 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0500011240.hg.1 | ENST00000282260.10 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0500011240.hg.1 | ENST00000282260.10 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0500011240.hg.1 | ENST00000334082.10 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0500011240.hg.1 | ENST00000334082.10 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0500011240.hg.1 | ENST00000334082.10 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0500011240.hg.1 | ENST00000334082.10 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0500011240.hg.1 | ENST00000334082.10 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0500011240.hg.1 | ENST00000334082.10 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0500011240.hg.1 | ENST00000334082.10 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0500011240.hg.1 | ENST00000334082.10 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0500011240.hg.1 | ENST00000334082.10 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0500011240.hg.1 | ENST00000334082.10 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0500011240.hg.1 | ENST00000334082.10 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0500011240.hg.1 | ENST00000334082.10 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0500011240.hg.1 | ENST00000334082.10 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0500011240.hg.1 | ENST00000334082.10 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0500011240.hg.1 | ENST00000334082.10 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0500011240.hg.1 | ENST00000460741.1 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0500011240.hg.1 | ENST00000460741.1 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0500011240.hg.1 | ENST00000460741.1 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0500011240.hg.1 | ENST00000460741.1 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0500011240.hg.1 | ENST00000460741.1 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0500011240.hg.1 | ENST00000508576.5 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0500011240.hg.1 | ENST00000508576.5 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0500011240.hg.1 | ENST00000508576.5 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0500011240.hg.1 | ENST00000508576.5 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0500011240.hg.1 | ENST00000508576.5 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0500011240.hg.1 | ENST00000508576.5 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0500011240.hg.1 | ENST00000535690.1 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0500011240.hg.1 | ENST00000535690.1 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0500011240.hg.1 | ENST00000535690.1 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0500011240.hg.1 | ENST00000535690.1 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0500011240.hg.1 | ENST00000535690.1 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0500011240.hg.1 | NM_001277077.1 | Refseq |
| Affymetrix Human Transcriptome Array 2.0 | TC0500011240.hg.1 | NM_001277077.1 | Refseq |
| Affymetrix Human Transcriptome Array 2.0 | TC0500011240.hg.1 | NM_001277077.1 | Refseq |
| Affymetrix Human Transcriptome Array 2.0 | TC0500011240.hg.1 | NM_001277077.1 | Refseq |
| Affymetrix Human Transcriptome Array 2.0 | TC0500011240.hg.1 | NM_001277077.1 | Refseq |
| Affymetrix Human Transcriptome Array 2.0 | TC0500011240.hg.1 | NM_001277077.1 | Refseq |
| Affymetrix Human Transcriptome Array 2.0 | TC0500011240.hg.1 | NM_001277077.1 | Refseq |
| Affymetrix Human Transcriptome Array 2.0 | TC0500011240.hg.1 | NM_001277077.1 | Refseq |
| Affymetrix Human Transcriptome Array 2.0 | TC0500011240.hg.1 | NM_001277077.1 | Refseq |
| Affymetrix Human Transcriptome Array 2.0 | TC0500011240.hg.1 | NM_001277077.1 | Refseq |
| Affymetrix Human Transcriptome Array 2.0 | TC0500011240.hg.1 | NM_001277078.1 | Refseq |
| Affymetrix Human Transcriptome Array 2.0 | TC0500011240.hg.1 | NM_001277078.1 | Refseq |
| Affymetrix Human Transcriptome Array 2.0 | TC0500011240.hg.1 | NM_001277078.1 | Refseq |
| Affymetrix Human Transcriptome Array 2.0 | TC0500011240.hg.1 | NM_001277078.1 | Refseq |
| Affymetrix Human Transcriptome Array 2.0 | TC0500011240.hg.1 | NM_001277078.1 | Refseq |
| Affymetrix Human Transcriptome Array 2.0 | TC0500011240.hg.1 | NM_001277078.1 | Refseq |
| Affymetrix Human Transcriptome Array 2.0 | TC0500011240.hg.1 | NM_001277078.1 | Refseq |
| Affymetrix Human Transcriptome Array 2.0 | TC0500011240.hg.1 | NM_001277078.1 | Refseq |
| Affymetrix Human Transcriptome Array 2.0 | TC0500011240.hg.1 | NM_001277078.1 | Refseq |
| Affymetrix Human Transcriptome Array 2.0 | TC0500011240.hg.1 | NM_001277078.1 | Refseq |
| Affymetrix Human Transcriptome Array 2.0 | TC0500011240.hg.1 | NM_004272.4 | Refseq |
| Affymetrix Human Transcriptome Array 2.0 | TC0500011240.hg.1 | NM_004272.4 | Refseq |
| Affymetrix Human Transcriptome Array 2.0 | TC0500011240.hg.1 | NM_004272.4 | Refseq |
| Affymetrix Human Transcriptome Array 2.0 | TC0500011240.hg.1 | NM_004272.4 | Refseq |
| Affymetrix Human Transcriptome Array 2.0 | TC0500011240.hg.1 | NM_004272.4 | Refseq |
| Affymetrix Human Transcriptome Array 2.0 | TC0500011240.hg.1 | NM_004272.4 | Refseq |
| Affymetrix Human Transcriptome Array 2.0 | TC0500011240.hg.1 | NM_004272.4 | Refseq |
| Affymetrix Human Transcriptome Array 2.0 | TC0500011240.hg.1 | NM_004272.4 | Refseq |
| Affymetrix Human Transcriptome Array 2.0 | TC0500011240.hg.1 | NM_004272.4 | Refseq |
| Affymetrix Human Transcriptome Array 2.0 | TC0500011240.hg.1 | NM_004272.4 | Refseq |
| Affymetrix Human Transcriptome Array 2.0 | TC0500011240.hg.1 | NM_004272.4 | Refseq |
| Affymetrix Human Transcriptome Array 2.0 | TC0500011240.hg.1 | NM_004272.4 | Refseq |
| Affymetrix Human Transcriptome Array 2.0 | TC0500011240.hg.1 | NM_004272.4 | Refseq |
| Affymetrix Human Transcriptome Array 2.0 | PSR05020875.hg.1 | TC05001522.hg.1 | HTA TC |
| Affymetrix Human Transcriptome Array 2.0 | PSR05020875.hg.1 | ENST00000334082.6 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | PSR05020875.hg.1 | NM_004272.4 | Refseq |
| Affymetrix Human Transcriptome Array 2.0 | PSR05020876.hg.1 | TC05001522.hg.1 | HTA TC |
| Affymetrix Human Transcriptome Array 2.0 | PSR05020876.hg.1 | ENST00000334082.6 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | PSR05020876.hg.1 | NM_004272.4 | Refseq |
| Affymetrix Human Transcriptome Array 2.0 | PSR05020873.hg.1 | TC05001522.hg.1 | HTA TC |
| Affymetrix Human Transcriptome Array 2.0 | PSR05020873.hg.1 | ENST00000282260.6 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | PSR05020873.hg.1 | ENST00000334082.6 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | PSR05020873.hg.1 | ENST00000508576.1 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | PSR05020873.hg.1 | NM_004272.4 | Refseq |
| Affymetrix Human Clariom D Assay | PSR0500178032.hg.1 | ENST00000282260.10 | Ensembl |
| Affymetrix Human Clariom D Assay | PSR0500178032.hg.1 | ENST00000334082.10 | Ensembl |
| Affymetrix Human Clariom D Assay | PSR0500178032.hg.1 | ENST00000508576.5 | Ensembl |
| Affymetrix Human Clariom D Assay | PSR0500178032.hg.1 | NM_001277077.1 | Refseq |
| Affymetrix Human Clariom D Assay | PSR0500178032.hg.1 | NM_001277078.1 | Refseq |
| Affymetrix Human Clariom D Assay | PSR0500178032.hg.1 | NM_004272.4 | Refseq |
| Affymetrix Human Clariom D Assay | PSR0500178032.hg.1 | ENST00000282260.10 | Ensembl |
| Affymetrix Human Clariom D Assay | PSR0500178032.hg.1 | ENST00000334082.10 | Ensembl |
| Affymetrix Human Clariom D Assay | PSR0500178032.hg.1 | ENST00000508576.5 | Ensembl |
| Affymetrix Human Clariom D Assay | PSR0500178032.hg.1 | NM_001277077.1 | Refseq |
| Affymetrix Human Clariom D Assay | PSR0500178032.hg.1 | NM_001277078.1 | Refseq |
| Affymetrix Human Clariom D Assay | PSR0500178032.hg.1 | NM_004272.4 | Refseq |
| Affymetrix Human Clariom D Assay | PSR0500178032.hg.1 | TC0500011240.hg.1 | Clariom D TC |
| Affymetrix Human Clariom D Assay | PSR0500178035.hg.1 | ENST00000334082.10 | Ensembl |
| Affymetrix Human Clariom D Assay | PSR0500178035.hg.1 | NM_001277077.1 | Refseq |
| Affymetrix Human Clariom D Assay | PSR0500178035.hg.1 | NM_001277078.1 | Refseq |
| Affymetrix Human Clariom D Assay | PSR0500178035.hg.1 | NM_004272.4 | Refseq |
| Affymetrix Human Clariom D Assay | PSR0500178035.hg.1 | ENST00000334082.10 | Ensembl |
| Affymetrix Human Clariom D Assay | PSR0500178035.hg.1 | NM_001277077.1 | Refseq |
| Affymetrix Human Clariom D Assay | PSR0500178035.hg.1 | NM_001277078.1 | Refseq |
| Affymetrix Human Clariom D Assay | PSR0500178035.hg.1 | NM_004272.4 | Refseq |
| Affymetrix Human Clariom D Assay | PSR0500178035.hg.1 | TC0500011240.hg.1 | Clariom D TC |
| Affymetrix Human Clariom D Assay | PSR0500178036.hg.1 | ENST00000334082.10 | Ensembl |
| Affymetrix Human Clariom D Assay | PSR0500178036.hg.1 | NM_001277077.1 | Refseq |
| Affymetrix Human Clariom D Assay | PSR0500178036.hg.1 | NM_001277078.1 | Refseq |
| Affymetrix Human Clariom D Assay | PSR0500178036.hg.1 | NM_004272.4 | Refseq |
| Affymetrix Human Clariom D Assay | PSR0500178036.hg.1 | ENST00000334082.10 | Ensembl |
| Affymetrix Human Clariom D Assay | PSR0500178036.hg.1 | NM_001277077.1 | Refseq |
| Affymetrix Human Clariom D Assay | PSR0500178036.hg.1 | NM_001277078.1 | Refseq |
| Affymetrix Human Clariom D Assay | PSR0500178036.hg.1 | NM_004272.4 | Refseq |
| Affymetrix Human Clariom D Assay | PSR0500178036.hg.1 | TC0500011240.hg.1 | Clariom D TC |
| Affymetrix Human Clariom D Assay | TC0500011240.hg.1 | ENST00000282260.10 | Ensembl |
| Affymetrix Human Clariom D Assay | TC0500011240.hg.1 | ENST00000282260.10 | Ensembl |
| Affymetrix Human Clariom D Assay | TC0500011240.hg.1 | ENST00000282260.10 | Ensembl |
| Affymetrix Human Clariom D Assay | TC0500011240.hg.1 | ENST00000282260.10 | Ensembl |
| Affymetrix Human Clariom D Assay | TC0500011240.hg.1 | ENST00000282260.10 | Ensembl |
| Affymetrix Human Clariom D Assay | TC0500011240.hg.1 | ENST00000282260.10 | Ensembl |
| Affymetrix Human Clariom D Assay | TC0500011240.hg.1 | ENST00000334082.10 | Ensembl |
| Affymetrix Human Clariom D Assay | TC0500011240.hg.1 | ENST00000334082.10 | Ensembl |
| Affymetrix Human Clariom D Assay | TC0500011240.hg.1 | ENST00000334082.10 | Ensembl |
| Affymetrix Human Clariom D Assay | TC0500011240.hg.1 | ENST00000334082.10 | Ensembl |
| Affymetrix Human Clariom D Assay | TC0500011240.hg.1 | ENST00000334082.10 | Ensembl |
| Affymetrix Human Clariom D Assay | TC0500011240.hg.1 | ENST00000334082.10 | Ensembl |
| Affymetrix Human Clariom D Assay | TC0500011240.hg.1 | ENST00000334082.10 | Ensembl |
| Affymetrix Human Clariom D Assay | TC0500011240.hg.1 | ENST00000334082.10 | Ensembl |
| Affymetrix Human Clariom D Assay | TC0500011240.hg.1 | ENST00000334082.10 | Ensembl |
| Affymetrix Human Clariom D Assay | TC0500011240.hg.1 | ENST00000334082.10 | Ensembl |
| Affymetrix Human Clariom D Assay | TC0500011240.hg.1 | ENST00000334082.10 | Ensembl |
| Affymetrix Human Clariom D Assay | TC0500011240.hg.1 | ENST00000334082.10 | Ensembl |
| Affymetrix Human Clariom D Assay | TC0500011240.hg.1 | ENST00000334082.10 | Ensembl |
| Affymetrix Human Clariom D Assay | TC0500011240.hg.1 | ENST00000334082.10 | Ensembl |
| Affymetrix Human Clariom D Assay | TC0500011240.hg.1 | ENST00000334082.10 | Ensembl |
| Affymetrix Human Clariom D Assay | TC0500011240.hg.1 | ENST00000460741.1 | Ensembl |
| Affymetrix Human Clariom D Assay | TC0500011240.hg.1 | ENST00000460741.1 | Ensembl |
| Affymetrix Human Clariom D Assay | TC0500011240.hg.1 | ENST00000460741.1 | Ensembl |
| Affymetrix Human Clariom D Assay | TC0500011240.hg.1 | ENST00000460741.1 | Ensembl |
| Affymetrix Human Clariom D Assay | TC0500011240.hg.1 | ENST00000460741.1 | Ensembl |
| Affymetrix Human Clariom D Assay | TC0500011240.hg.1 | ENST00000508576.5 | Ensembl |
| Affymetrix Human Clariom D Assay | TC0500011240.hg.1 | ENST00000508576.5 | Ensembl |
| Affymetrix Human Clariom D Assay | TC0500011240.hg.1 | ENST00000508576.5 | Ensembl |
| Affymetrix Human Clariom D Assay | TC0500011240.hg.1 | ENST00000508576.5 | Ensembl |
| Affymetrix Human Clariom D Assay | TC0500011240.hg.1 | ENST00000508576.5 | Ensembl |
| Affymetrix Human Clariom D Assay | TC0500011240.hg.1 | ENST00000508576.5 | Ensembl |
| Affymetrix Human Clariom D Assay | TC0500011240.hg.1 | ENST00000535690.1 | Ensembl |
| Affymetrix Human Clariom D Assay | TC0500011240.hg.1 | ENST00000535690.1 | Ensembl |
| Affymetrix Human Clariom D Assay | TC0500011240.hg.1 | ENST00000535690.1 | Ensembl |
| Affymetrix Human Clariom D Assay | TC0500011240.hg.1 | ENST00000535690.1 | Ensembl |
| Affymetrix Human Clariom D Assay | TC0500011240.hg.1 | ENST00000535690.1 | Ensembl |
| Affymetrix Human Clariom D Assay | TC0500011240.hg.1 | NM_001277077.1 | Refseq |
| Affymetrix Human Clariom D Assay | TC0500011240.hg.1 | NM_001277077.1 | Refseq |
| Affymetrix Human Clariom D Assay | TC0500011240.hg.1 | NM_001277077.1 | Refseq |
| Affymetrix Human Clariom D Assay | TC0500011240.hg.1 | NM_001277077.1 | Refseq |
| Affymetrix Human Clariom D Assay | TC0500011240.hg.1 | NM_001277077.1 | Refseq |
| Affymetrix Human Clariom D Assay | TC0500011240.hg.1 | NM_001277077.1 | Refseq |
| Affymetrix Human Clariom D Assay | TC0500011240.hg.1 | NM_001277077.1 | Refseq |
| Affymetrix Human Clariom D Assay | TC0500011240.hg.1 | NM_001277077.1 | Refseq |
| Affymetrix Human Clariom D Assay | TC0500011240.hg.1 | NM_001277077.1 | Refseq |
| Affymetrix Human Clariom D Assay | TC0500011240.hg.1 | NM_001277077.1 | Refseq |
| Affymetrix Human Clariom D Assay | TC0500011240.hg.1 | NM_001277078.1 | Refseq |
| Affymetrix Human Clariom D Assay | TC0500011240.hg.1 | NM_001277078.1 | Refseq |
| Affymetrix Human Clariom D Assay | TC0500011240.hg.1 | NM_001277078.1 | Refseq |
| Affymetrix Human Clariom D Assay | TC0500011240.hg.1 | NM_001277078.1 | Refseq |
| Affymetrix Human Clariom D Assay | TC0500011240.hg.1 | NM_001277078.1 | Refseq |
| Affymetrix Human Clariom D Assay | TC0500011240.hg.1 | NM_001277078.1 | Refseq |
| Affymetrix Human Clariom D Assay | TC0500011240.hg.1 | NM_001277078.1 | Refseq |
| Affymetrix Human Clariom D Assay | TC0500011240.hg.1 | NM_001277078.1 | Refseq |
| Affymetrix Human Clariom D Assay | TC0500011240.hg.1 | NM_001277078.1 | Refseq |
| Affymetrix Human Clariom D Assay | TC0500011240.hg.1 | NM_001277078.1 | Refseq |
| Affymetrix Human Clariom D Assay | TC0500011240.hg.1 | NM_004272.4 | Refseq |
| Affymetrix Human Clariom D Assay | TC0500011240.hg.1 | NM_004272.4 | Refseq |
| Affymetrix Human Clariom D Assay | TC0500011240.hg.1 | NM_004272.4 | Refseq |
| Affymetrix Human Clariom D Assay | TC0500011240.hg.1 | NM_004272.4 | Refseq |
| Affymetrix Human Clariom D Assay | TC0500011240.hg.1 | NM_004272.4 | Refseq |
| Affymetrix Human Clariom D Assay | TC0500011240.hg.1 | NM_004272.4 | Refseq |
| Affymetrix Human Clariom D Assay | TC0500011240.hg.1 | NM_004272.4 | Refseq |
| Affymetrix Human Clariom D Assay | TC0500011240.hg.1 | NM_004272.4 | Refseq |
| Affymetrix Human Clariom D Assay | TC0500011240.hg.1 | NM_004272.4 | Refseq |
| Affymetrix Human Clariom D Assay | TC0500011240.hg.1 | NM_004272.4 | Refseq |
| Affymetrix Human Clariom D Assay | TC0500011240.hg.1 | NM_004272.4 | Refseq |
| Affymetrix Human Clariom D Assay | TC0500011240.hg.1 | NM_004272.4 | Refseq |
| Affymetrix Human Clariom D Assay | TC0500011240.hg.1 | NM_004272.4 | Refseq |
| Array Name | Probe IDs | Targets | Target Type |
|---|---|---|---|
| ABI Human Genome Survey Microarray Version 2.0 | 104128 | - | - |

HOMER1
Genomic Map