Assay Details
| Gene Symbol: PNPLA4 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | NM_001142389.1 | NP_001135861.1 | 3 - 4 | 347 | 66 | |
| NM_001172672.1 | NP_001166143.1 | 2 - 3 | 154 | 66 | ||
| NM_004650.2 | NP_004641.1 | 3 - 4 | 442 | 66 | ||
| XM_011545588.1 | XP_011543890.1 | 3 - 4 | 1210 | 66 | ||
| GenBank mRNA | AK289888.1 | - | 3 - 4 | 392 | 66 | |
| AK304586.1 | - | 2 - 3 | 139 | 66 | ||
| BC020746.1 | - | 3 - 4 | 452 | 4717901 | 66 | |
| BM850121.1 | - | 3 - 4 | 362 | 66 | ||
| DC421932.1 | - | 3 - 4 | 363 | 66 | ||
| U03886.1 | - | 3 - 4 | 410 | 66 | ||
More Information
Updated Mapping Information
This assay targets the same location as HTA transcript cluster(s) TC0X000827.hg.1
Additional Information:
This assay is in the 5'UTR region of the AK304586.1,NM_001172672.1 transcripts.
Set Membership: |
|
Matching Ambion siRNAs:
Panther Classification:
Gene Ontology Categories:
MicroArray Probes:
Affymetrix
| Array Name | Probe IDs | Targets | Target Type |
|---|---|---|---|
| Affymetrix Human Transcriptome Array 2.0 | PSR0X011253.hg.1 | TC0X000827.hg.1 | HTA TC |
| Affymetrix Human Transcriptome Array 2.0 | PSR0X011253.hg.1 | ENST00000381042.4 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | PSR0X011253.hg.1 | ENST00000444736.1 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | PSR0X011253.hg.1 | ENST00000537427.1 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | PSR0X011253.hg.1 | NM_001142389.1 | Refseq |
| Affymetrix Human Transcriptome Array 2.0 | PSR0X011253.hg.1 | NM_001172672.1 | Refseq |
| Affymetrix Human Transcriptome Array 2.0 | PSR0X011253.hg.1 | NM_004650.2 | Refseq |
| Affymetrix Human Transcriptome Array 2.0 | PSR0X011260.hg.1 | TC0X000827.hg.1 | HTA TC |
| Affymetrix Human Transcriptome Array 2.0 | PSR0X011260.hg.1 | ENST00000444736.1 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | PSR0X011260.hg.1 | ENST00000537427.1 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | PSR0X011260.hg.1 | NM_001142389.1 | Refseq |
| Affymetrix Human Transcriptome Array 2.0 | PSR0X011260.hg.1 | NM_001172672.1 | Refseq |
| Affymetrix Human Transcriptome Array 2.0 | PSR0X011255.hg.1 | TC0X000827.hg.1 | HTA TC |
| Affymetrix Human Transcriptome Array 2.0 | PSR0X011255.hg.1 | ENST00000381042.4 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | PSR0X011255.hg.1 | ENST00000444736.1 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | PSR0X011255.hg.1 | NM_001142389.1 | Refseq |
| Affymetrix Human Transcriptome Array 2.0 | PSR0X011255.hg.1 | NM_004650.2 | Refseq |
| Affymetrix Human Transcriptome Array 2.0 | TC0X00009018.hg.1 | ENST00000381042.8 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0X00009018.hg.1 | ENST00000381042.8 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0X00009018.hg.1 | ENST00000381042.8 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0X00009018.hg.1 | ENST00000381042.8 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0X00009018.hg.1 | ENST00000381042.8 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0X00009018.hg.1 | ENST00000381042.8 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0X00009018.hg.1 | ENST00000381042.8 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0X00009018.hg.1 | ENST00000381042.8 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0X00009018.hg.1 | ENST00000381042.8 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0X00009018.hg.1 | ENST00000381042.8 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0X00009018.hg.1 | ENST00000442940.1 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0X00009018.hg.1 | ENST00000442940.1 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0X00009018.hg.1 | ENST00000442940.1 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0X00009018.hg.1 | ENST00000442940.1 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0X00009018.hg.1 | ENST00000442940.1 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0X00009018.hg.1 | ENST00000442940.1 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0X00009018.hg.1 | ENST00000444736.5 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0X00009018.hg.1 | ENST00000444736.5 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0X00009018.hg.1 | ENST00000444736.5 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0X00009018.hg.1 | ENST00000444736.5 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0X00009018.hg.1 | ENST00000444736.5 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0X00009018.hg.1 | ENST00000444736.5 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0X00009018.hg.1 | ENST00000444736.5 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0X00009018.hg.1 | ENST00000444736.5 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0X00009018.hg.1 | ENST00000444736.5 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0X00009018.hg.1 | ENST00000537427.5 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0X00009018.hg.1 | ENST00000537427.5 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0X00009018.hg.1 | ENST00000537427.5 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0X00009018.hg.1 | ENST00000537427.5 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0X00009018.hg.1 | ENST00000537427.5 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0X00009018.hg.1 | ENST00000537427.5 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0X00009018.hg.1 | ENST00000537427.5 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0X00009018.hg.1 | ENST00000537427.5 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0X00009018.hg.1 | NM_001142389.1 | Refseq |
| Affymetrix Human Transcriptome Array 2.0 | TC0X00009018.hg.1 | NM_001142389.1 | Refseq |
| Affymetrix Human Transcriptome Array 2.0 | TC0X00009018.hg.1 | NM_001142389.1 | Refseq |
| Affymetrix Human Transcriptome Array 2.0 | TC0X00009018.hg.1 | NM_001142389.1 | Refseq |
| Affymetrix Human Transcriptome Array 2.0 | TC0X00009018.hg.1 | NM_001142389.1 | Refseq |
| Affymetrix Human Transcriptome Array 2.0 | TC0X00009018.hg.1 | NM_001142389.1 | Refseq |
| Affymetrix Human Transcriptome Array 2.0 | TC0X00009018.hg.1 | NM_001142389.1 | Refseq |
| Affymetrix Human Transcriptome Array 2.0 | TC0X00009018.hg.1 | NM_001142389.1 | Refseq |
| Affymetrix Human Transcriptome Array 2.0 | TC0X00009018.hg.1 | NM_001142389.1 | Refseq |
| Affymetrix Human Transcriptome Array 2.0 | TC0X00009018.hg.1 | NM_001172672.1 | Refseq |
| Affymetrix Human Transcriptome Array 2.0 | TC0X00009018.hg.1 | NM_001172672.1 | Refseq |
| Affymetrix Human Transcriptome Array 2.0 | TC0X00009018.hg.1 | NM_001172672.1 | Refseq |
| Affymetrix Human Transcriptome Array 2.0 | TC0X00009018.hg.1 | NM_001172672.1 | Refseq |
| Affymetrix Human Transcriptome Array 2.0 | TC0X00009018.hg.1 | NM_001172672.1 | Refseq |
| Affymetrix Human Transcriptome Array 2.0 | TC0X00009018.hg.1 | NM_001172672.1 | Refseq |
| Affymetrix Human Transcriptome Array 2.0 | TC0X00009018.hg.1 | NM_001172672.1 | Refseq |
| Affymetrix Human Transcriptome Array 2.0 | TC0X00009018.hg.1 | NM_001172672.1 | Refseq |
| Affymetrix Human Transcriptome Array 2.0 | TC0X00009018.hg.1 | NM_004650.2 | Refseq |
| Affymetrix Human Transcriptome Array 2.0 | TC0X00009018.hg.1 | NM_004650.2 | Refseq |
| Affymetrix Human Transcriptome Array 2.0 | TC0X00009018.hg.1 | NM_004650.2 | Refseq |
| Affymetrix Human Transcriptome Array 2.0 | TC0X00009018.hg.1 | NM_004650.2 | Refseq |
| Affymetrix Human Transcriptome Array 2.0 | TC0X00009018.hg.1 | NM_004650.2 | Refseq |
| Affymetrix Human Transcriptome Array 2.0 | TC0X00009018.hg.1 | NM_004650.2 | Refseq |
| Affymetrix Human Transcriptome Array 2.0 | TC0X00009018.hg.1 | NM_004650.2 | Refseq |
| Affymetrix Human Transcriptome Array 2.0 | TC0X00009018.hg.1 | NM_004650.2 | Refseq |
| Affymetrix Human Transcriptome Array 2.0 | TC0X00009018.hg.1 | NM_004650.2 | Refseq |
| Affymetrix Human Transcriptome Array 2.0 | PSR0X011254.hg.1 | TC0X000827.hg.1 | HTA TC |
| Affymetrix Human Transcriptome Array 2.0 | PSR0X011254.hg.1 | ENST00000381042.4 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | PSR0X011254.hg.1 | ENST00000444736.1 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | PSR0X011254.hg.1 | ENST00000537427.1 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | PSR0X011254.hg.1 | NM_001142389.1 | Refseq |
| Affymetrix Human Transcriptome Array 2.0 | PSR0X011254.hg.1 | NM_001172672.1 | Refseq |
| Affymetrix Human Transcriptome Array 2.0 | PSR0X011254.hg.1 | NM_004650.2 | Refseq |
| Affymetrix Human Transcriptome Array 2.0 | TC0Y00006649.hg.1 | ENST00000458328.1 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0Y00006649.hg.1 | ENST00000458328.1 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0Y00006649.hg.1 | ENST00000458328.1 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | PSR0X011256.hg.1 | TC0X000827.hg.1 | HTA TC |
| Affymetrix Human Transcriptome Array 2.0 | PSR0X011256.hg.1 | ENST00000381042.4 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | PSR0X011256.hg.1 | NM_004650.2 | Refseq |
| Affymetrix Human Clariom D Assay | PSR0X00161861.hg.1 | ENST00000442940.1 | Ensembl |
| Affymetrix Human Clariom D Assay | PSR0X00161861.hg.1 | ENST00000444736.5 | Ensembl |
| Affymetrix Human Clariom D Assay | PSR0X00161861.hg.1 | ENST00000537427.5 | Ensembl |
| Affymetrix Human Clariom D Assay | PSR0X00161861.hg.1 | NM_001142389.1 | Refseq |
| Affymetrix Human Clariom D Assay | PSR0X00161861.hg.1 | NM_001172672.1 | Refseq |
| Affymetrix Human Clariom D Assay | PSR0X00161861.hg.1 | ENST00000442940.1 | Ensembl |
| Affymetrix Human Clariom D Assay | PSR0X00161861.hg.1 | ENST00000444736.5 | Ensembl |
| Affymetrix Human Clariom D Assay | PSR0X00161861.hg.1 | ENST00000537427.5 | Ensembl |
| Affymetrix Human Clariom D Assay | PSR0X00161861.hg.1 | NM_001142389.1 | Refseq |
| Affymetrix Human Clariom D Assay | PSR0X00161861.hg.1 | NM_001172672.1 | Refseq |
| Affymetrix Human Clariom D Assay | PSR0X00161861.hg.1 | TC0X00009018.hg.1 | Clariom D TC |
| Affymetrix Human Clariom D Assay | PSR0X00161854.hg.1 | ENST00000381042.8 | Ensembl |
| Affymetrix Human Clariom D Assay | PSR0X00161854.hg.1 | ENST00000442940.1 | Ensembl |
| Affymetrix Human Clariom D Assay | PSR0X00161854.hg.1 | ENST00000444736.5 | Ensembl |
| Affymetrix Human Clariom D Assay | PSR0X00161854.hg.1 | ENST00000537427.5 | Ensembl |
| Affymetrix Human Clariom D Assay | PSR0X00161854.hg.1 | NM_001142389.1 | Refseq |
| Affymetrix Human Clariom D Assay | PSR0X00161854.hg.1 | NM_001172672.1 | Refseq |
| Affymetrix Human Clariom D Assay | PSR0X00161854.hg.1 | NM_004650.2 | Refseq |
| Affymetrix Human Clariom D Assay | PSR0X00161854.hg.1 | ENST00000381042.8 | Ensembl |
| Affymetrix Human Clariom D Assay | PSR0X00161854.hg.1 | ENST00000442940.1 | Ensembl |
| Affymetrix Human Clariom D Assay | PSR0X00161854.hg.1 | ENST00000444736.5 | Ensembl |
| Affymetrix Human Clariom D Assay | PSR0X00161854.hg.1 | ENST00000537427.5 | Ensembl |
| Affymetrix Human Clariom D Assay | PSR0X00161854.hg.1 | NM_001142389.1 | Refseq |
| Affymetrix Human Clariom D Assay | PSR0X00161854.hg.1 | NM_001172672.1 | Refseq |
| Affymetrix Human Clariom D Assay | PSR0X00161854.hg.1 | NM_004650.2 | Refseq |
| Affymetrix Human Clariom D Assay | PSR0X00161854.hg.1 | TC0X00009018.hg.1 | Clariom D TC |
| Affymetrix Human Clariom D Assay | PSR0X00161855.hg.1 | ENST00000381042.8 | Ensembl |
| Affymetrix Human Clariom D Assay | PSR0X00161855.hg.1 | ENST00000442940.1 | Ensembl |
| Affymetrix Human Clariom D Assay | PSR0X00161855.hg.1 | ENST00000444736.5 | Ensembl |
| Affymetrix Human Clariom D Assay | PSR0X00161855.hg.1 | NM_001142389.1 | Refseq |
| Affymetrix Human Clariom D Assay | PSR0X00161855.hg.1 | NM_004650.2 | Refseq |
| Affymetrix Human Clariom D Assay | PSR0X00161855.hg.1 | ENST00000381042.8 | Ensembl |
| Affymetrix Human Clariom D Assay | PSR0X00161855.hg.1 | ENST00000442940.1 | Ensembl |
| Affymetrix Human Clariom D Assay | PSR0X00161855.hg.1 | ENST00000444736.5 | Ensembl |
| Affymetrix Human Clariom D Assay | PSR0X00161855.hg.1 | NM_001142389.1 | Refseq |
| Affymetrix Human Clariom D Assay | PSR0X00161855.hg.1 | NM_004650.2 | Refseq |
| Affymetrix Human Clariom D Assay | PSR0X00161855.hg.1 | TC0X00009018.hg.1 | Clariom D TC |
| Affymetrix Human Clariom D Assay | TC0Y00006649.hg.1 | ENST00000458328.1 | Ensembl |
| Affymetrix Human Clariom D Assay | TC0Y00006649.hg.1 | ENST00000458328.1 | Ensembl |
| Affymetrix Human Clariom D Assay | TC0Y00006649.hg.1 | ENST00000458328.1 | Ensembl |
| Affymetrix Human Clariom D Assay | PSR0X00161852.hg.1 | ENST00000381042.8 | Ensembl |
| Affymetrix Human Clariom D Assay | PSR0X00161852.hg.1 | ENST00000442940.1 | Ensembl |
| Affymetrix Human Clariom D Assay | PSR0X00161852.hg.1 | ENST00000444736.5 | Ensembl |
| Affymetrix Human Clariom D Assay | PSR0X00161852.hg.1 | ENST00000537427.5 | Ensembl |
| Affymetrix Human Clariom D Assay | PSR0X00161852.hg.1 | NM_001142389.1 | Refseq |
| Affymetrix Human Clariom D Assay | PSR0X00161852.hg.1 | NM_001172672.1 | Refseq |
| Affymetrix Human Clariom D Assay | PSR0X00161852.hg.1 | NM_004650.2 | Refseq |
| Affymetrix Human Clariom D Assay | PSR0X00161852.hg.1 | ENST00000381042.8 | Ensembl |
| Affymetrix Human Clariom D Assay | PSR0X00161852.hg.1 | ENST00000442940.1 | Ensembl |
| Affymetrix Human Clariom D Assay | PSR0X00161852.hg.1 | ENST00000444736.5 | Ensembl |
| Affymetrix Human Clariom D Assay | PSR0X00161852.hg.1 | ENST00000537427.5 | Ensembl |
| Affymetrix Human Clariom D Assay | PSR0X00161852.hg.1 | NM_001142389.1 | Refseq |
| Affymetrix Human Clariom D Assay | PSR0X00161852.hg.1 | NM_001172672.1 | Refseq |
| Affymetrix Human Clariom D Assay | PSR0X00161852.hg.1 | NM_004650.2 | Refseq |
| Affymetrix Human Clariom D Assay | PSR0X00161852.hg.1 | TC0X00009018.hg.1 | Clariom D TC |
| Affymetrix Human Clariom D Assay | TC0X00009018.hg.1 | ENST00000381042.8 | Ensembl |
| Affymetrix Human Clariom D Assay | TC0X00009018.hg.1 | ENST00000381042.8 | Ensembl |
| Affymetrix Human Clariom D Assay | TC0X00009018.hg.1 | ENST00000381042.8 | Ensembl |
| Affymetrix Human Clariom D Assay | TC0X00009018.hg.1 | ENST00000381042.8 | Ensembl |
| Affymetrix Human Clariom D Assay | TC0X00009018.hg.1 | ENST00000381042.8 | Ensembl |
| Affymetrix Human Clariom D Assay | TC0X00009018.hg.1 | ENST00000381042.8 | Ensembl |
| Affymetrix Human Clariom D Assay | TC0X00009018.hg.1 | ENST00000381042.8 | Ensembl |
| Affymetrix Human Clariom D Assay | TC0X00009018.hg.1 | ENST00000381042.8 | Ensembl |
| Affymetrix Human Clariom D Assay | TC0X00009018.hg.1 | ENST00000381042.8 | Ensembl |
| Affymetrix Human Clariom D Assay | TC0X00009018.hg.1 | ENST00000381042.8 | Ensembl |
| Affymetrix Human Clariom D Assay | TC0X00009018.hg.1 | ENST00000442940.1 | Ensembl |
| Affymetrix Human Clariom D Assay | TC0X00009018.hg.1 | ENST00000442940.1 | Ensembl |
| Affymetrix Human Clariom D Assay | TC0X00009018.hg.1 | ENST00000442940.1 | Ensembl |
| Affymetrix Human Clariom D Assay | TC0X00009018.hg.1 | ENST00000442940.1 | Ensembl |
| Affymetrix Human Clariom D Assay | TC0X00009018.hg.1 | ENST00000442940.1 | Ensembl |
| Affymetrix Human Clariom D Assay | TC0X00009018.hg.1 | ENST00000442940.1 | Ensembl |
| Affymetrix Human Clariom D Assay | TC0X00009018.hg.1 | ENST00000444736.5 | Ensembl |
| Affymetrix Human Clariom D Assay | TC0X00009018.hg.1 | ENST00000444736.5 | Ensembl |
| Affymetrix Human Clariom D Assay | TC0X00009018.hg.1 | ENST00000444736.5 | Ensembl |
| Affymetrix Human Clariom D Assay | TC0X00009018.hg.1 | ENST00000444736.5 | Ensembl |
| Affymetrix Human Clariom D Assay | TC0X00009018.hg.1 | ENST00000444736.5 | Ensembl |
| Affymetrix Human Clariom D Assay | TC0X00009018.hg.1 | ENST00000444736.5 | Ensembl |
| Affymetrix Human Clariom D Assay | TC0X00009018.hg.1 | ENST00000444736.5 | Ensembl |
| Affymetrix Human Clariom D Assay | TC0X00009018.hg.1 | ENST00000444736.5 | Ensembl |
| Affymetrix Human Clariom D Assay | TC0X00009018.hg.1 | ENST00000444736.5 | Ensembl |
| Affymetrix Human Clariom D Assay | TC0X00009018.hg.1 | ENST00000537427.5 | Ensembl |
| Affymetrix Human Clariom D Assay | TC0X00009018.hg.1 | ENST00000537427.5 | Ensembl |
| Affymetrix Human Clariom D Assay | TC0X00009018.hg.1 | ENST00000537427.5 | Ensembl |
| Affymetrix Human Clariom D Assay | TC0X00009018.hg.1 | ENST00000537427.5 | Ensembl |
| Affymetrix Human Clariom D Assay | TC0X00009018.hg.1 | ENST00000537427.5 | Ensembl |
| Affymetrix Human Clariom D Assay | TC0X00009018.hg.1 | ENST00000537427.5 | Ensembl |
| Affymetrix Human Clariom D Assay | TC0X00009018.hg.1 | ENST00000537427.5 | Ensembl |
| Affymetrix Human Clariom D Assay | TC0X00009018.hg.1 | ENST00000537427.5 | Ensembl |
| Affymetrix Human Clariom D Assay | TC0X00009018.hg.1 | NM_001142389.1 | Refseq |
| Affymetrix Human Clariom D Assay | TC0X00009018.hg.1 | NM_001142389.1 | Refseq |
| Affymetrix Human Clariom D Assay | TC0X00009018.hg.1 | NM_001142389.1 | Refseq |
| Affymetrix Human Clariom D Assay | TC0X00009018.hg.1 | NM_001142389.1 | Refseq |
| Affymetrix Human Clariom D Assay | TC0X00009018.hg.1 | NM_001142389.1 | Refseq |
| Affymetrix Human Clariom D Assay | TC0X00009018.hg.1 | NM_001142389.1 | Refseq |
| Affymetrix Human Clariom D Assay | TC0X00009018.hg.1 | NM_001142389.1 | Refseq |
| Affymetrix Human Clariom D Assay | TC0X00009018.hg.1 | NM_001142389.1 | Refseq |
| Affymetrix Human Clariom D Assay | TC0X00009018.hg.1 | NM_001142389.1 | Refseq |
| Affymetrix Human Clariom D Assay | TC0X00009018.hg.1 | NM_001172672.1 | Refseq |
| Affymetrix Human Clariom D Assay | TC0X00009018.hg.1 | NM_001172672.1 | Refseq |
| Affymetrix Human Clariom D Assay | TC0X00009018.hg.1 | NM_001172672.1 | Refseq |
| Affymetrix Human Clariom D Assay | TC0X00009018.hg.1 | NM_001172672.1 | Refseq |
| Affymetrix Human Clariom D Assay | TC0X00009018.hg.1 | NM_001172672.1 | Refseq |
| Affymetrix Human Clariom D Assay | TC0X00009018.hg.1 | NM_001172672.1 | Refseq |
| Affymetrix Human Clariom D Assay | TC0X00009018.hg.1 | NM_001172672.1 | Refseq |
| Affymetrix Human Clariom D Assay | TC0X00009018.hg.1 | NM_001172672.1 | Refseq |
| Affymetrix Human Clariom D Assay | TC0X00009018.hg.1 | NM_004650.2 | Refseq |
| Affymetrix Human Clariom D Assay | TC0X00009018.hg.1 | NM_004650.2 | Refseq |
| Affymetrix Human Clariom D Assay | TC0X00009018.hg.1 | NM_004650.2 | Refseq |
| Affymetrix Human Clariom D Assay | TC0X00009018.hg.1 | NM_004650.2 | Refseq |
| Affymetrix Human Clariom D Assay | TC0X00009018.hg.1 | NM_004650.2 | Refseq |
| Affymetrix Human Clariom D Assay | TC0X00009018.hg.1 | NM_004650.2 | Refseq |
| Affymetrix Human Clariom D Assay | TC0X00009018.hg.1 | NM_004650.2 | Refseq |
| Affymetrix Human Clariom D Assay | TC0X00009018.hg.1 | NM_004650.2 | Refseq |
| Affymetrix Human Clariom D Assay | TC0X00009018.hg.1 | NM_004650.2 | Refseq |
| Affymetrix Human Clariom D Assay | PSR0X00161857.hg.1 | ENST00000381042.8 | Ensembl |
| Affymetrix Human Clariom D Assay | PSR0X00161857.hg.1 | NM_004650.2 | Refseq |
| Affymetrix Human Clariom D Assay | PSR0X00161857.hg.1 | ENST00000381042.8 | Ensembl |
| Affymetrix Human Clariom D Assay | PSR0X00161857.hg.1 | NM_004650.2 | Refseq |
| Affymetrix Human Clariom D Assay | PSR0X00161857.hg.1 | TC0X00009018.hg.1 | Clariom D TC |

PNPLA4
Genomic Map