Assay Details
| Gene Symbol: ENSA | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | NM_207042.1 | NP_996925.1 | 3 - 4 | 375 | 118 | |
| NM_207043.1 | NP_996926.1 | 3 - 4 | 375 | 118 | ||
| NM_207045.1 | NP_996928.1 | 3 - 4 | 239 | 118 | ||
| GenBank mRNA | BU858280.1 | - | 233 | 118 | ||
More Information
Set Membership: |
|

ENSA
Genomic Map