Assay Details
| Gene Symbol: SEC31A | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | NM_001077206.3 | NP_001070674.1 | 15 - 16 | 1895 | 146 | |
| NM_001077207.3 | NP_001070675.1 | 15 - 16 | 1895 | 146 | ||
| NM_001077208.3 | NP_001070676.1 | 15 - 16 | 1895 | 146 | ||
| NM_001191049.2 | NP_001177978.1 | 14 - 15 | 1812 | 146 | ||
| NM_001318119.1 | NP_001305048.1 | 15 - 16 | 1892 | 146 | ||
| NM_001318120.1 | NP_001305049.1 | 17 - 18 | 2109 | 146 | ||
| GenBank mRNA | AB018359.1 | - | 15 - 16 | 1751 | 146 | |
| AB020712.1 | - | 17 - 18 | 2076 | 146 | ||
| AF139184.1 | - | 14 - 15 | 1698 | 146 | ||
| AK001014.1 | - | 4 - 5 | 208 | 146 | ||
| AK002161.1 | - | 15 - 16 | 1768 | 146 | ||
| AK295810.1 | - | 14 - 15 | 1710 | 146 | ||
| BC084583.1 | - | 15 - 16 | 1734 | 6196402 | 146 | |
| BC117221.1 | - | 15 - 16 | 1763 | 40125772 | 146 | |
| BC143489.1 | - | 15 - 16 | 1760 | 9051997 | 146 | |
| BC143491.1 | - | 15 - 16 | 1763 | 9051999 | 146 | |
More Information
Set Membership: |
|

SEC31A
Genomic Map