Assay Details
| Gene Symbol: ABCF2 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | XM_017011639.1 | XP_016867128.1 | 1 - 2 | 59 | 60 | |
More Information
Additional Information:
This assay is in the 5'UTR region of the XM_017011639.1 transcripts.
Set Membership: |
|
Panther Classification:
Gene Ontology Categories:
MicroArray Probes:
Affymetrix
| Array Name | Probe IDs | Targets | Target Type |
|---|---|---|---|
| Affymetrix Human Transcriptome Array 2.0 | TC0700013065.hg.1 | ENST00000222388.6 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0700013065.hg.1 | ENST00000222388.6 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0700013065.hg.1 | ENST00000222388.6 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0700013065.hg.1 | ENST00000222388.6 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0700013065.hg.1 | ENST00000222388.6 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0700013065.hg.1 | ENST00000222388.6 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0700013065.hg.1 | ENST00000222388.6 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0700013065.hg.1 | ENST00000222388.6 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0700013065.hg.1 | ENST00000222388.6 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0700013065.hg.1 | ENST00000222388.6 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0700013065.hg.1 | ENST00000222388.6 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0700013065.hg.1 | ENST00000222388.6 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0700013065.hg.1 | ENST00000222388.6 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0700013065.hg.1 | ENST00000222388.6 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0700013065.hg.1 | ENST00000222388.6 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0700013065.hg.1 | ENST00000222388.6 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0700013065.hg.1 | ENST00000222388.6 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0700013065.hg.1 | ENST00000222388.6 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0700013065.hg.1 | ENST00000222388.6 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0700013065.hg.1 | ENST00000287844.6 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0700013065.hg.1 | ENST00000287844.6 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0700013065.hg.1 | ENST00000287844.6 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0700013065.hg.1 | ENST00000287844.6 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0700013065.hg.1 | ENST00000287844.6 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0700013065.hg.1 | ENST00000287844.6 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0700013065.hg.1 | ENST00000287844.6 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0700013065.hg.1 | ENST00000287844.6 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0700013065.hg.1 | ENST00000287844.6 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0700013065.hg.1 | ENST00000287844.6 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0700013065.hg.1 | ENST00000287844.6 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0700013065.hg.1 | ENST00000287844.6 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0700013065.hg.1 | ENST00000287844.6 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0700013065.hg.1 | ENST00000287844.6 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0700013065.hg.1 | ENST00000287844.6 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0700013065.hg.1 | ENST00000287844.6 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0700013065.hg.1 | ENST00000287844.6 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0700013065.hg.1 | ENST00000287844.6 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0700013065.hg.1 | ENST00000287844.6 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0700013065.hg.1 | ENST00000287844.6 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0700013065.hg.1 | ENST00000287844.6 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0700013065.hg.1 | ENST00000441774.1 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0700013065.hg.1 | ENST00000441774.1 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0700013065.hg.1 | ENST00000441774.1 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0700013065.hg.1 | ENST00000441774.1 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0700013065.hg.1 | ENST00000441774.1 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0700013065.hg.1 | ENST00000441774.1 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0700013065.hg.1 | ENST00000441774.1 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0700013065.hg.1 | ENST00000468073.5 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0700013065.hg.1 | ENST00000468073.5 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0700013065.hg.1 | ENST00000468073.5 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0700013065.hg.1 | ENST00000468073.5 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0700013065.hg.1 | ENST00000468073.5 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0700013065.hg.1 | ENST00000468073.5 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0700013065.hg.1 | ENST00000468073.5 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0700013065.hg.1 | ENST00000468073.5 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0700013065.hg.1 | ENST00000473874.1 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0700013065.hg.1 | ENST00000473874.1 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0700013065.hg.1 | ENST00000473874.1 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0700013065.hg.1 | ENST00000473874.1 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0700013065.hg.1 | ENST00000473874.1 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0700013065.hg.1 | ENST00000473874.1 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0700013065.hg.1 | ENST00000473874.1 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0700013065.hg.1 | ENST00000477252.1 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0700013065.hg.1 | ENST00000477252.1 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0700013065.hg.1 | ENST00000477252.1 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0700013065.hg.1 | NM_005692.4 | Refseq |
| Affymetrix Human Transcriptome Array 2.0 | TC0700013065.hg.1 | NM_005692.4 | Refseq |
| Affymetrix Human Transcriptome Array 2.0 | TC0700013065.hg.1 | NM_005692.4 | Refseq |
| Affymetrix Human Transcriptome Array 2.0 | TC0700013065.hg.1 | NM_005692.4 | Refseq |
| Affymetrix Human Transcriptome Array 2.0 | TC0700013065.hg.1 | NM_005692.4 | Refseq |
| Affymetrix Human Transcriptome Array 2.0 | TC0700013065.hg.1 | NM_005692.4 | Refseq |
| Affymetrix Human Transcriptome Array 2.0 | TC0700013065.hg.1 | NM_005692.4 | Refseq |
| Affymetrix Human Transcriptome Array 2.0 | TC0700013065.hg.1 | NM_005692.4 | Refseq |
| Affymetrix Human Transcriptome Array 2.0 | TC0700013065.hg.1 | NM_005692.4 | Refseq |
| Affymetrix Human Transcriptome Array 2.0 | TC0700013065.hg.1 | NM_005692.4 | Refseq |
| Affymetrix Human Transcriptome Array 2.0 | TC0700013065.hg.1 | NM_005692.4 | Refseq |
| Affymetrix Human Transcriptome Array 2.0 | TC0700013065.hg.1 | NM_005692.4 | Refseq |
| Affymetrix Human Transcriptome Array 2.0 | TC0700013065.hg.1 | NM_005692.4 | Refseq |
| Affymetrix Human Transcriptome Array 2.0 | TC0700013065.hg.1 | NM_005692.4 | Refseq |
| Affymetrix Human Transcriptome Array 2.0 | TC0700013065.hg.1 | NM_005692.4 | Refseq |
| Affymetrix Human Transcriptome Array 2.0 | TC0700013065.hg.1 | NM_005692.4 | Refseq |
| Affymetrix Human Transcriptome Array 2.0 | TC0700013065.hg.1 | NM_005692.4 | Refseq |
| Affymetrix Human Transcriptome Array 2.0 | TC0700013065.hg.1 | NM_005692.4 | Refseq |
| Affymetrix Human Transcriptome Array 2.0 | TC0700013065.hg.1 | NM_005692.4 | Refseq |
| Affymetrix Human Transcriptome Array 2.0 | TC0700013065.hg.1 | NM_005692.4 | Refseq |
| Affymetrix Human Transcriptome Array 2.0 | TC0700013065.hg.1 | NM_005692.4 | Refseq |
| Affymetrix Human Transcriptome Array 2.0 | TC0700013065.hg.1 | NM_005692.4 | Refseq |
| Affymetrix Human Transcriptome Array 2.0 | TC0700013065.hg.1 | NM_007189.2 | Refseq |
| Affymetrix Human Transcriptome Array 2.0 | TC0700013065.hg.1 | NM_007189.2 | Refseq |
| Affymetrix Human Transcriptome Array 2.0 | TC0700013065.hg.1 | NM_007189.2 | Refseq |
| Affymetrix Human Transcriptome Array 2.0 | TC0700013065.hg.1 | NM_007189.2 | Refseq |
| Affymetrix Human Transcriptome Array 2.0 | TC0700013065.hg.1 | NM_007189.2 | Refseq |
| Affymetrix Human Transcriptome Array 2.0 | TC0700013065.hg.1 | NM_007189.2 | Refseq |
| Affymetrix Human Transcriptome Array 2.0 | TC0700013065.hg.1 | NM_007189.2 | Refseq |
| Affymetrix Human Transcriptome Array 2.0 | TC0700013065.hg.1 | NM_007189.2 | Refseq |
| Affymetrix Human Transcriptome Array 2.0 | TC0700013065.hg.1 | NM_007189.2 | Refseq |
| Affymetrix Human Transcriptome Array 2.0 | TC0700013065.hg.1 | NM_007189.2 | Refseq |
| Affymetrix Human Transcriptome Array 2.0 | TC0700013065.hg.1 | NM_007189.2 | Refseq |
| Affymetrix Human Transcriptome Array 2.0 | TC0700013065.hg.1 | NM_007189.2 | Refseq |
| Affymetrix Human Transcriptome Array 2.0 | TC0700013065.hg.1 | NM_007189.2 | Refseq |
| Affymetrix Human Transcriptome Array 2.0 | TC0700013065.hg.1 | NM_007189.2 | Refseq |
| Affymetrix Human Transcriptome Array 2.0 | TC0700013065.hg.1 | NM_007189.2 | Refseq |
| Affymetrix Human Transcriptome Array 2.0 | TC0700013065.hg.1 | NM_007189.2 | Refseq |
| Affymetrix Human Transcriptome Array 2.0 | TC0700013065.hg.1 | NM_007189.2 | Refseq |
| Affymetrix Human Transcriptome Array 2.0 | TC0700013065.hg.1 | NM_007189.2 | Refseq |
| Affymetrix Human Transcriptome Array 2.0 | TC0700013065.hg.1 | NM_007189.2 | Refseq |
| Affymetrix Human Transcriptome Array 2.0 | TC0700013065.hg.1 | NM_007189.2 | Refseq |
| Affymetrix Human Transcriptome Array 2.0 | TC0700013065.hg.1 | NM_007189.2 | Refseq |
| Affymetrix Human Transcriptome Array 2.0 | TC0700013065.hg.1 | NM_007189.2 | Refseq |
| Affymetrix Human Transcriptome Array 2.0 | TC0700013065.hg.1 | NM_007189.2 | Refseq |
| Affymetrix Human Clariom D Assay | TC0700013065.hg.1 | ENST00000222388.6 | Ensembl |
| Affymetrix Human Clariom D Assay | TC0700013065.hg.1 | ENST00000222388.6 | Ensembl |
| Affymetrix Human Clariom D Assay | TC0700013065.hg.1 | ENST00000222388.6 | Ensembl |
| Affymetrix Human Clariom D Assay | TC0700013065.hg.1 | ENST00000222388.6 | Ensembl |
| Affymetrix Human Clariom D Assay | TC0700013065.hg.1 | ENST00000222388.6 | Ensembl |
| Affymetrix Human Clariom D Assay | TC0700013065.hg.1 | ENST00000222388.6 | Ensembl |
| Affymetrix Human Clariom D Assay | TC0700013065.hg.1 | ENST00000222388.6 | Ensembl |
| Affymetrix Human Clariom D Assay | TC0700013065.hg.1 | ENST00000222388.6 | Ensembl |
| Affymetrix Human Clariom D Assay | TC0700013065.hg.1 | ENST00000222388.6 | Ensembl |
| Affymetrix Human Clariom D Assay | TC0700013065.hg.1 | ENST00000222388.6 | Ensembl |
| Affymetrix Human Clariom D Assay | TC0700013065.hg.1 | ENST00000222388.6 | Ensembl |
| Affymetrix Human Clariom D Assay | TC0700013065.hg.1 | ENST00000222388.6 | Ensembl |
| Affymetrix Human Clariom D Assay | TC0700013065.hg.1 | ENST00000222388.6 | Ensembl |
| Affymetrix Human Clariom D Assay | TC0700013065.hg.1 | ENST00000222388.6 | Ensembl |
| Affymetrix Human Clariom D Assay | TC0700013065.hg.1 | ENST00000222388.6 | Ensembl |
| Affymetrix Human Clariom D Assay | TC0700013065.hg.1 | ENST00000222388.6 | Ensembl |
| Affymetrix Human Clariom D Assay | TC0700013065.hg.1 | ENST00000222388.6 | Ensembl |
| Affymetrix Human Clariom D Assay | TC0700013065.hg.1 | ENST00000222388.6 | Ensembl |
| Affymetrix Human Clariom D Assay | TC0700013065.hg.1 | ENST00000222388.6 | Ensembl |
| Affymetrix Human Clariom D Assay | TC0700013065.hg.1 | ENST00000287844.6 | Ensembl |
| Affymetrix Human Clariom D Assay | TC0700013065.hg.1 | ENST00000287844.6 | Ensembl |
| Affymetrix Human Clariom D Assay | TC0700013065.hg.1 | ENST00000287844.6 | Ensembl |
| Affymetrix Human Clariom D Assay | TC0700013065.hg.1 | ENST00000287844.6 | Ensembl |
| Affymetrix Human Clariom D Assay | TC0700013065.hg.1 | ENST00000287844.6 | Ensembl |
| Affymetrix Human Clariom D Assay | TC0700013065.hg.1 | ENST00000287844.6 | Ensembl |
| Affymetrix Human Clariom D Assay | TC0700013065.hg.1 | ENST00000287844.6 | Ensembl |
| Affymetrix Human Clariom D Assay | TC0700013065.hg.1 | ENST00000287844.6 | Ensembl |
| Affymetrix Human Clariom D Assay | TC0700013065.hg.1 | ENST00000287844.6 | Ensembl |
| Affymetrix Human Clariom D Assay | TC0700013065.hg.1 | ENST00000287844.6 | Ensembl |
| Affymetrix Human Clariom D Assay | TC0700013065.hg.1 | ENST00000287844.6 | Ensembl |
| Affymetrix Human Clariom D Assay | TC0700013065.hg.1 | ENST00000287844.6 | Ensembl |
| Affymetrix Human Clariom D Assay | TC0700013065.hg.1 | ENST00000287844.6 | Ensembl |
| Affymetrix Human Clariom D Assay | TC0700013065.hg.1 | ENST00000287844.6 | Ensembl |
| Affymetrix Human Clariom D Assay | TC0700013065.hg.1 | ENST00000287844.6 | Ensembl |
| Affymetrix Human Clariom D Assay | TC0700013065.hg.1 | ENST00000287844.6 | Ensembl |
| Affymetrix Human Clariom D Assay | TC0700013065.hg.1 | ENST00000287844.6 | Ensembl |
| Affymetrix Human Clariom D Assay | TC0700013065.hg.1 | ENST00000287844.6 | Ensembl |
| Affymetrix Human Clariom D Assay | TC0700013065.hg.1 | ENST00000287844.6 | Ensembl |
| Affymetrix Human Clariom D Assay | TC0700013065.hg.1 | ENST00000287844.6 | Ensembl |
| Affymetrix Human Clariom D Assay | TC0700013065.hg.1 | ENST00000287844.6 | Ensembl |
| Affymetrix Human Clariom D Assay | TC0700013065.hg.1 | ENST00000441774.1 | Ensembl |
| Affymetrix Human Clariom D Assay | TC0700013065.hg.1 | ENST00000441774.1 | Ensembl |
| Affymetrix Human Clariom D Assay | TC0700013065.hg.1 | ENST00000441774.1 | Ensembl |
| Affymetrix Human Clariom D Assay | TC0700013065.hg.1 | ENST00000441774.1 | Ensembl |
| Affymetrix Human Clariom D Assay | TC0700013065.hg.1 | ENST00000441774.1 | Ensembl |
| Affymetrix Human Clariom D Assay | TC0700013065.hg.1 | ENST00000441774.1 | Ensembl |
| Affymetrix Human Clariom D Assay | TC0700013065.hg.1 | ENST00000441774.1 | Ensembl |
| Affymetrix Human Clariom D Assay | TC0700013065.hg.1 | ENST00000468073.5 | Ensembl |
| Affymetrix Human Clariom D Assay | TC0700013065.hg.1 | ENST00000468073.5 | Ensembl |
| Affymetrix Human Clariom D Assay | TC0700013065.hg.1 | ENST00000468073.5 | Ensembl |
| Affymetrix Human Clariom D Assay | TC0700013065.hg.1 | ENST00000468073.5 | Ensembl |
| Affymetrix Human Clariom D Assay | TC0700013065.hg.1 | ENST00000468073.5 | Ensembl |
| Affymetrix Human Clariom D Assay | TC0700013065.hg.1 | ENST00000468073.5 | Ensembl |
| Affymetrix Human Clariom D Assay | TC0700013065.hg.1 | ENST00000468073.5 | Ensembl |
| Affymetrix Human Clariom D Assay | TC0700013065.hg.1 | ENST00000468073.5 | Ensembl |
| Affymetrix Human Clariom D Assay | TC0700013065.hg.1 | ENST00000473874.1 | Ensembl |
| Affymetrix Human Clariom D Assay | TC0700013065.hg.1 | ENST00000473874.1 | Ensembl |
| Affymetrix Human Clariom D Assay | TC0700013065.hg.1 | ENST00000473874.1 | Ensembl |
| Affymetrix Human Clariom D Assay | TC0700013065.hg.1 | ENST00000473874.1 | Ensembl |
| Affymetrix Human Clariom D Assay | TC0700013065.hg.1 | ENST00000473874.1 | Ensembl |
| Affymetrix Human Clariom D Assay | TC0700013065.hg.1 | ENST00000473874.1 | Ensembl |
| Affymetrix Human Clariom D Assay | TC0700013065.hg.1 | ENST00000473874.1 | Ensembl |
| Affymetrix Human Clariom D Assay | TC0700013065.hg.1 | ENST00000477252.1 | Ensembl |
| Affymetrix Human Clariom D Assay | TC0700013065.hg.1 | ENST00000477252.1 | Ensembl |
| Affymetrix Human Clariom D Assay | TC0700013065.hg.1 | ENST00000477252.1 | Ensembl |
| Affymetrix Human Clariom D Assay | TC0700013065.hg.1 | NM_005692.4 | Refseq |
| Affymetrix Human Clariom D Assay | TC0700013065.hg.1 | NM_005692.4 | Refseq |
| Affymetrix Human Clariom D Assay | TC0700013065.hg.1 | NM_005692.4 | Refseq |
| Affymetrix Human Clariom D Assay | TC0700013065.hg.1 | NM_005692.4 | Refseq |
| Affymetrix Human Clariom D Assay | TC0700013065.hg.1 | NM_005692.4 | Refseq |
| Affymetrix Human Clariom D Assay | TC0700013065.hg.1 | NM_005692.4 | Refseq |
| Affymetrix Human Clariom D Assay | TC0700013065.hg.1 | NM_005692.4 | Refseq |
| Affymetrix Human Clariom D Assay | TC0700013065.hg.1 | NM_005692.4 | Refseq |
| Affymetrix Human Clariom D Assay | TC0700013065.hg.1 | NM_005692.4 | Refseq |
| Affymetrix Human Clariom D Assay | TC0700013065.hg.1 | NM_005692.4 | Refseq |
| Affymetrix Human Clariom D Assay | TC0700013065.hg.1 | NM_005692.4 | Refseq |
| Affymetrix Human Clariom D Assay | TC0700013065.hg.1 | NM_005692.4 | Refseq |
| Affymetrix Human Clariom D Assay | TC0700013065.hg.1 | NM_005692.4 | Refseq |
| Affymetrix Human Clariom D Assay | TC0700013065.hg.1 | NM_005692.4 | Refseq |
| Affymetrix Human Clariom D Assay | TC0700013065.hg.1 | NM_005692.4 | Refseq |
| Affymetrix Human Clariom D Assay | TC0700013065.hg.1 | NM_005692.4 | Refseq |
| Affymetrix Human Clariom D Assay | TC0700013065.hg.1 | NM_005692.4 | Refseq |
| Affymetrix Human Clariom D Assay | TC0700013065.hg.1 | NM_005692.4 | Refseq |
| Affymetrix Human Clariom D Assay | TC0700013065.hg.1 | NM_005692.4 | Refseq |
| Affymetrix Human Clariom D Assay | TC0700013065.hg.1 | NM_005692.4 | Refseq |
| Affymetrix Human Clariom D Assay | TC0700013065.hg.1 | NM_005692.4 | Refseq |
| Affymetrix Human Clariom D Assay | TC0700013065.hg.1 | NM_005692.4 | Refseq |
| Affymetrix Human Clariom D Assay | TC0700013065.hg.1 | NM_007189.2 | Refseq |
| Affymetrix Human Clariom D Assay | TC0700013065.hg.1 | NM_007189.2 | Refseq |
| Affymetrix Human Clariom D Assay | TC0700013065.hg.1 | NM_007189.2 | Refseq |
| Affymetrix Human Clariom D Assay | TC0700013065.hg.1 | NM_007189.2 | Refseq |
| Affymetrix Human Clariom D Assay | TC0700013065.hg.1 | NM_007189.2 | Refseq |
| Affymetrix Human Clariom D Assay | TC0700013065.hg.1 | NM_007189.2 | Refseq |
| Affymetrix Human Clariom D Assay | TC0700013065.hg.1 | NM_007189.2 | Refseq |
| Affymetrix Human Clariom D Assay | TC0700013065.hg.1 | NM_007189.2 | Refseq |
| Affymetrix Human Clariom D Assay | TC0700013065.hg.1 | NM_007189.2 | Refseq |
| Affymetrix Human Clariom D Assay | TC0700013065.hg.1 | NM_007189.2 | Refseq |
| Affymetrix Human Clariom D Assay | TC0700013065.hg.1 | NM_007189.2 | Refseq |
| Affymetrix Human Clariom D Assay | TC0700013065.hg.1 | NM_007189.2 | Refseq |
| Affymetrix Human Clariom D Assay | TC0700013065.hg.1 | NM_007189.2 | Refseq |
| Affymetrix Human Clariom D Assay | TC0700013065.hg.1 | NM_007189.2 | Refseq |
| Affymetrix Human Clariom D Assay | TC0700013065.hg.1 | NM_007189.2 | Refseq |
| Affymetrix Human Clariom D Assay | TC0700013065.hg.1 | NM_007189.2 | Refseq |
| Affymetrix Human Clariom D Assay | TC0700013065.hg.1 | NM_007189.2 | Refseq |
| Affymetrix Human Clariom D Assay | TC0700013065.hg.1 | NM_007189.2 | Refseq |
| Affymetrix Human Clariom D Assay | TC0700013065.hg.1 | NM_007189.2 | Refseq |
| Affymetrix Human Clariom D Assay | TC0700013065.hg.1 | NM_007189.2 | Refseq |
| Affymetrix Human Clariom D Assay | TC0700013065.hg.1 | NM_007189.2 | Refseq |
| Affymetrix Human Clariom D Assay | TC0700013065.hg.1 | NM_007189.2 | Refseq |
| Affymetrix Human Clariom D Assay | TC0700013065.hg.1 | NM_007189.2 | Refseq |

ABCF2
Genomic Map