Assay Details
| Gene Symbol: LRP10 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | XM_005267510.1 | XP_005267567.1 | 2 - 3 | 771 | 106 | |
| GenBank mRNA | AK291030.1 | - | 2 - 3 | 225 | 106 | |
| AL834518.1 | - | 2 - 3 | 230 | 106 | ||
| AY358399.1 | - | 2 - 3 | 625 | 106 | ||
| BC113714.1 | - | 2 - 3 | 107 | 8322766 | 106 | |
| BC113716.1 | - | 2 - 3 | 107 | 8322768 | 106 | |
| BX161500.1 | - | 2 - 3 | 553 | 106 | ||
More Information
Updated Mapping Information
Set Membership: |
|
Panther Classification:
Gene Ontology Categories:
MicroArray Probes:
Affymetrix
ABI
| Array Name | Probe IDs | Targets | Target Type |
|---|---|---|---|
| Affymetrix Human Transcriptome Array 2.0 | TC1400006663.hg.1 | ENST00000359591.8 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC1400006663.hg.1 | ENST00000359591.8 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC1400006663.hg.1 | ENST00000359591.8 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC1400006663.hg.1 | ENST00000359591.8 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC1400006663.hg.1 | ENST00000359591.8 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC1400006663.hg.1 | ENST00000359591.8 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC1400006663.hg.1 | ENST00000359591.8 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC1400006663.hg.1 | ENST00000359591.8 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC1400006663.hg.1 | ENST00000359591.8 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC1400006663.hg.1 | ENST00000359591.8 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC1400006663.hg.1 | ENST00000359591.8 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC1400006663.hg.1 | ENST00000359591.8 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC1400006663.hg.1 | ENST00000359591.8 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC1400006663.hg.1 | ENST00000359591.8 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC1400006663.hg.1 | ENST00000359591.8 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC1400006663.hg.1 | ENST00000359591.8 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC1400006663.hg.1 | ENST00000359591.8 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC1400006663.hg.1 | ENST00000359591.8 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC1400006663.hg.1 | ENST00000359591.8 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC1400006663.hg.1 | ENST00000359591.8 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC1400006663.hg.1 | ENST00000359591.8 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC1400006663.hg.1 | ENST00000359591.8 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC1400006663.hg.1 | ENST00000470660.1 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC1400006663.hg.1 | ENST00000470660.1 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC1400006663.hg.1 | ENST00000470660.1 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC1400006663.hg.1 | ENST00000470660.1 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC1400006663.hg.1 | ENST00000470660.1 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC1400006663.hg.1 | ENST00000546834.5 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC1400006663.hg.1 | ENST00000546834.5 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC1400006663.hg.1 | ENST00000546834.5 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC1400006663.hg.1 | ENST00000546834.5 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC1400006663.hg.1 | ENST00000546834.5 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC1400006663.hg.1 | ENST00000546834.5 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC1400006663.hg.1 | ENST00000546834.5 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC1400006663.hg.1 | ENST00000546834.5 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC1400006663.hg.1 | ENST00000546834.5 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC1400006663.hg.1 | ENST00000546834.5 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC1400006663.hg.1 | ENST00000546834.5 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC1400006663.hg.1 | ENST00000551466.1 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC1400006663.hg.1 | ENST00000551466.1 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC1400006663.hg.1 | ENST00000551466.1 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC1400006663.hg.1 | ENST00000551466.1 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC1400006663.hg.1 | ENST00000551466.1 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC1400006663.hg.1 | ENST00000551466.1 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC1400006663.hg.1 | ENST00000551466.1 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC1400006663.hg.1 | ENST00000551466.1 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC1400006663.hg.1 | ENST00000553002.1 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC1400006663.hg.1 | ENST00000553002.1 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC1400006663.hg.1 | ENST00000553002.1 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC1400006663.hg.1 | ENST00000553002.1 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC1400006663.hg.1 | ENST00000553002.1 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC1400006663.hg.1 | NM_014045.4 | Refseq |
| Affymetrix Human Transcriptome Array 2.0 | TC1400006663.hg.1 | NM_014045.4 | Refseq |
| Affymetrix Human Transcriptome Array 2.0 | TC1400006663.hg.1 | NM_014045.4 | Refseq |
| Affymetrix Human Transcriptome Array 2.0 | TC1400006663.hg.1 | NM_014045.4 | Refseq |
| Affymetrix Human Transcriptome Array 2.0 | TC1400006663.hg.1 | NM_014045.4 | Refseq |
| Affymetrix Human Transcriptome Array 2.0 | TC1400006663.hg.1 | NM_014045.4 | Refseq |
| Affymetrix Human Transcriptome Array 2.0 | TC1400006663.hg.1 | NM_014045.4 | Refseq |
| Affymetrix Human Transcriptome Array 2.0 | TC1400006663.hg.1 | NM_014045.4 | Refseq |
| Affymetrix Human Transcriptome Array 2.0 | TC1400006663.hg.1 | NM_014045.4 | Refseq |
| Affymetrix Human Transcriptome Array 2.0 | TC1400006663.hg.1 | NM_014045.4 | Refseq |
| Affymetrix Human Transcriptome Array 2.0 | TC1400006663.hg.1 | NM_014045.4 | Refseq |
| Affymetrix Human Transcriptome Array 2.0 | TC1400006663.hg.1 | NM_014045.4 | Refseq |
| Affymetrix Human Transcriptome Array 2.0 | TC1400006663.hg.1 | NM_014045.4 | Refseq |
| Affymetrix Human Transcriptome Array 2.0 | TC1400006663.hg.1 | NM_014045.4 | Refseq |
| Affymetrix Human Transcriptome Array 2.0 | TC1400006663.hg.1 | NM_014045.4 | Refseq |
| Affymetrix Human Transcriptome Array 2.0 | TC1400006663.hg.1 | NM_014045.4 | Refseq |
| Affymetrix Human Transcriptome Array 2.0 | TC1400006663.hg.1 | NM_014045.4 | Refseq |
| Affymetrix Human Clariom D Assay | PSR1400145548.hg.1 | ENST00000359591.8 | Ensembl |
| Affymetrix Human Clariom D Assay | PSR1400145548.hg.1 | ENST00000546834.5 | Ensembl |
| Affymetrix Human Clariom D Assay | PSR1400145548.hg.1 | NM_014045.4 | Refseq |
| Affymetrix Human Clariom D Assay | PSR1400145548.hg.1 | ENST00000359591.8 | Ensembl |
| Affymetrix Human Clariom D Assay | PSR1400145548.hg.1 | ENST00000546834.5 | Ensembl |
| Affymetrix Human Clariom D Assay | PSR1400145548.hg.1 | NM_014045.4 | Refseq |
| Affymetrix Human Clariom D Assay | PSR1400145548.hg.1 | TC1400006663.hg.1 | Clariom D TC |
| Affymetrix Human Clariom D Assay | TC1400006663.hg.1 | ENST00000359591.8 | Ensembl |
| Affymetrix Human Clariom D Assay | TC1400006663.hg.1 | ENST00000359591.8 | Ensembl |
| Affymetrix Human Clariom D Assay | TC1400006663.hg.1 | ENST00000359591.8 | Ensembl |
| Affymetrix Human Clariom D Assay | TC1400006663.hg.1 | ENST00000359591.8 | Ensembl |
| Affymetrix Human Clariom D Assay | TC1400006663.hg.1 | ENST00000359591.8 | Ensembl |
| Affymetrix Human Clariom D Assay | TC1400006663.hg.1 | ENST00000359591.8 | Ensembl |
| Affymetrix Human Clariom D Assay | TC1400006663.hg.1 | ENST00000359591.8 | Ensembl |
| Affymetrix Human Clariom D Assay | TC1400006663.hg.1 | ENST00000359591.8 | Ensembl |
| Affymetrix Human Clariom D Assay | TC1400006663.hg.1 | ENST00000359591.8 | Ensembl |
| Affymetrix Human Clariom D Assay | TC1400006663.hg.1 | ENST00000359591.8 | Ensembl |
| Affymetrix Human Clariom D Assay | TC1400006663.hg.1 | ENST00000359591.8 | Ensembl |
| Affymetrix Human Clariom D Assay | TC1400006663.hg.1 | ENST00000359591.8 | Ensembl |
| Affymetrix Human Clariom D Assay | TC1400006663.hg.1 | ENST00000359591.8 | Ensembl |
| Affymetrix Human Clariom D Assay | TC1400006663.hg.1 | ENST00000359591.8 | Ensembl |
| Affymetrix Human Clariom D Assay | TC1400006663.hg.1 | ENST00000359591.8 | Ensembl |
| Affymetrix Human Clariom D Assay | TC1400006663.hg.1 | ENST00000359591.8 | Ensembl |
| Affymetrix Human Clariom D Assay | TC1400006663.hg.1 | ENST00000359591.8 | Ensembl |
| Affymetrix Human Clariom D Assay | TC1400006663.hg.1 | ENST00000359591.8 | Ensembl |
| Affymetrix Human Clariom D Assay | TC1400006663.hg.1 | ENST00000359591.8 | Ensembl |
| Affymetrix Human Clariom D Assay | TC1400006663.hg.1 | ENST00000359591.8 | Ensembl |
| Affymetrix Human Clariom D Assay | TC1400006663.hg.1 | ENST00000359591.8 | Ensembl |
| Affymetrix Human Clariom D Assay | TC1400006663.hg.1 | ENST00000359591.8 | Ensembl |
| Affymetrix Human Clariom D Assay | TC1400006663.hg.1 | ENST00000470660.1 | Ensembl |
| Affymetrix Human Clariom D Assay | TC1400006663.hg.1 | ENST00000470660.1 | Ensembl |
| Affymetrix Human Clariom D Assay | TC1400006663.hg.1 | ENST00000470660.1 | Ensembl |
| Affymetrix Human Clariom D Assay | TC1400006663.hg.1 | ENST00000470660.1 | Ensembl |
| Affymetrix Human Clariom D Assay | TC1400006663.hg.1 | ENST00000470660.1 | Ensembl |
| Affymetrix Human Clariom D Assay | TC1400006663.hg.1 | ENST00000546834.5 | Ensembl |
| Affymetrix Human Clariom D Assay | TC1400006663.hg.1 | ENST00000546834.5 | Ensembl |
| Affymetrix Human Clariom D Assay | TC1400006663.hg.1 | ENST00000546834.5 | Ensembl |
| Affymetrix Human Clariom D Assay | TC1400006663.hg.1 | ENST00000546834.5 | Ensembl |
| Affymetrix Human Clariom D Assay | TC1400006663.hg.1 | ENST00000546834.5 | Ensembl |
| Affymetrix Human Clariom D Assay | TC1400006663.hg.1 | ENST00000546834.5 | Ensembl |
| Affymetrix Human Clariom D Assay | TC1400006663.hg.1 | ENST00000546834.5 | Ensembl |
| Affymetrix Human Clariom D Assay | TC1400006663.hg.1 | ENST00000546834.5 | Ensembl |
| Affymetrix Human Clariom D Assay | TC1400006663.hg.1 | ENST00000546834.5 | Ensembl |
| Affymetrix Human Clariom D Assay | TC1400006663.hg.1 | ENST00000546834.5 | Ensembl |
| Affymetrix Human Clariom D Assay | TC1400006663.hg.1 | ENST00000546834.5 | Ensembl |
| Affymetrix Human Clariom D Assay | TC1400006663.hg.1 | ENST00000551466.1 | Ensembl |
| Affymetrix Human Clariom D Assay | TC1400006663.hg.1 | ENST00000551466.1 | Ensembl |
| Affymetrix Human Clariom D Assay | TC1400006663.hg.1 | ENST00000551466.1 | Ensembl |
| Affymetrix Human Clariom D Assay | TC1400006663.hg.1 | ENST00000551466.1 | Ensembl |
| Affymetrix Human Clariom D Assay | TC1400006663.hg.1 | ENST00000551466.1 | Ensembl |
| Affymetrix Human Clariom D Assay | TC1400006663.hg.1 | ENST00000551466.1 | Ensembl |
| Affymetrix Human Clariom D Assay | TC1400006663.hg.1 | ENST00000551466.1 | Ensembl |
| Affymetrix Human Clariom D Assay | TC1400006663.hg.1 | ENST00000551466.1 | Ensembl |
| Affymetrix Human Clariom D Assay | TC1400006663.hg.1 | ENST00000553002.1 | Ensembl |
| Affymetrix Human Clariom D Assay | TC1400006663.hg.1 | ENST00000553002.1 | Ensembl |
| Affymetrix Human Clariom D Assay | TC1400006663.hg.1 | ENST00000553002.1 | Ensembl |
| Affymetrix Human Clariom D Assay | TC1400006663.hg.1 | ENST00000553002.1 | Ensembl |
| Affymetrix Human Clariom D Assay | TC1400006663.hg.1 | ENST00000553002.1 | Ensembl |
| Affymetrix Human Clariom D Assay | TC1400006663.hg.1 | NM_014045.4 | Refseq |
| Affymetrix Human Clariom D Assay | TC1400006663.hg.1 | NM_014045.4 | Refseq |
| Affymetrix Human Clariom D Assay | TC1400006663.hg.1 | NM_014045.4 | Refseq |
| Affymetrix Human Clariom D Assay | TC1400006663.hg.1 | NM_014045.4 | Refseq |
| Affymetrix Human Clariom D Assay | TC1400006663.hg.1 | NM_014045.4 | Refseq |
| Affymetrix Human Clariom D Assay | TC1400006663.hg.1 | NM_014045.4 | Refseq |
| Affymetrix Human Clariom D Assay | TC1400006663.hg.1 | NM_014045.4 | Refseq |
| Affymetrix Human Clariom D Assay | TC1400006663.hg.1 | NM_014045.4 | Refseq |
| Affymetrix Human Clariom D Assay | TC1400006663.hg.1 | NM_014045.4 | Refseq |
| Affymetrix Human Clariom D Assay | TC1400006663.hg.1 | NM_014045.4 | Refseq |
| Affymetrix Human Clariom D Assay | TC1400006663.hg.1 | NM_014045.4 | Refseq |
| Affymetrix Human Clariom D Assay | TC1400006663.hg.1 | NM_014045.4 | Refseq |
| Affymetrix Human Clariom D Assay | TC1400006663.hg.1 | NM_014045.4 | Refseq |
| Affymetrix Human Clariom D Assay | TC1400006663.hg.1 | NM_014045.4 | Refseq |
| Affymetrix Human Clariom D Assay | TC1400006663.hg.1 | NM_014045.4 | Refseq |
| Affymetrix Human Clariom D Assay | TC1400006663.hg.1 | NM_014045.4 | Refseq |
| Affymetrix Human Clariom D Assay | TC1400006663.hg.1 | NM_014045.4 | Refseq |
| Affymetrix Human Clariom D Assay | PSR1400145546.hg.1 | ENST00000359591.8 | Ensembl |
| Affymetrix Human Clariom D Assay | PSR1400145546.hg.1 | NM_014045.4 | Refseq |
| Affymetrix Human Clariom D Assay | PSR1400145546.hg.1 | ENST00000359591.8 | Ensembl |
| Affymetrix Human Clariom D Assay | PSR1400145546.hg.1 | NM_014045.4 | Refseq |
| Affymetrix Human Clariom D Assay | PSR1400145546.hg.1 | TC1400006663.hg.1 | Clariom D TC |
| Affymetrix Human Clariom D Assay | PSR1400145547.hg.1 | ENST00000359591.8 | Ensembl |
| Affymetrix Human Clariom D Assay | PSR1400145547.hg.1 | ENST00000546834.5 | Ensembl |
| Affymetrix Human Clariom D Assay | PSR1400145547.hg.1 | NM_014045.4 | Refseq |
| Affymetrix Human Clariom D Assay | PSR1400145547.hg.1 | ENST00000359591.8 | Ensembl |
| Affymetrix Human Clariom D Assay | PSR1400145547.hg.1 | ENST00000546834.5 | Ensembl |
| Affymetrix Human Clariom D Assay | PSR1400145547.hg.1 | NM_014045.4 | Refseq |
| Affymetrix Human Clariom D Assay | PSR1400145547.hg.1 | TC1400006663.hg.1 | Clariom D TC |
| Affymetrix Human Clariom D Assay | PSR1400145550.hg.1 | ENST00000359591.8 | Ensembl |
| Affymetrix Human Clariom D Assay | PSR1400145550.hg.1 | ENST00000546834.5 | Ensembl |
| Affymetrix Human Clariom D Assay | PSR1400145550.hg.1 | ENST00000553002.1 | Ensembl |
| Affymetrix Human Clariom D Assay | PSR1400145550.hg.1 | NM_014045.4 | Refseq |
| Affymetrix Human Clariom D Assay | PSR1400145550.hg.1 | ENST00000359591.8 | Ensembl |
| Affymetrix Human Clariom D Assay | PSR1400145550.hg.1 | ENST00000546834.5 | Ensembl |
| Affymetrix Human Clariom D Assay | PSR1400145550.hg.1 | ENST00000553002.1 | Ensembl |
| Affymetrix Human Clariom D Assay | PSR1400145550.hg.1 | NM_014045.4 | Refseq |
| Affymetrix Human Clariom D Assay | PSR1400145550.hg.1 | TC1400006663.hg.1 | Clariom D TC |
| Array Name | Probe IDs | Targets | Target Type |
|---|---|---|---|
| ABI Human Genome Survey Microarray Version 2.0 | 178639 | - | - |
| ABI Human Genome Survey Microarray Version 2.0 | 210403 | - | - |

LRP10
Genomic Map