Assay Details
| Gene Symbol: COX11 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | XM_017024193.1 | XP_016879682.1 | 4 - 5 | 4825 | 82 | |
| XM_017024196.1 | XP_016879685.1 | 4 - 5 | 978 | 82 | ||
More Information
Set Membership: |
|
Panther Classification:
Gene Ontology Categories:
MicroArray Probes:
Affymetrix
| Array Name | Probe IDs | Targets | Target Type |
|---|---|---|---|
| Affymetrix Human Transcriptome Array 2.0 | TC1700011171.hg.1 | ENST00000299335.7 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC1700011171.hg.1 | ENST00000299335.7 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC1700011171.hg.1 | ENST00000299335.7 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC1700011171.hg.1 | ENST00000299335.7 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC1700011171.hg.1 | ENST00000299335.7 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC1700011171.hg.1 | ENST00000299335.7 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC1700011171.hg.1 | ENST00000299335.7 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC1700011171.hg.1 | ENST00000299335.7 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC1700011171.hg.1 | ENST00000299335.7 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC1700011171.hg.1 | ENST00000299335.7 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC1700011171.hg.1 | ENST00000571584.1 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC1700011171.hg.1 | ENST00000571584.1 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC1700011171.hg.1 | ENST00000571584.1 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC1700011171.hg.1 | ENST00000571584.1 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC1700011171.hg.1 | ENST00000571584.1 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC1700011171.hg.1 | ENST00000572088.1 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC1700011171.hg.1 | ENST00000572088.1 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC1700011171.hg.1 | ENST00000572088.1 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC1700011171.hg.1 | ENST00000572088.1 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC1700011171.hg.1 | ENST00000572088.1 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC1700011171.hg.1 | ENST00000572088.1 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC1700011171.hg.1 | ENST00000572558.5 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC1700011171.hg.1 | ENST00000572558.5 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC1700011171.hg.1 | ENST00000572558.5 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC1700011171.hg.1 | ENST00000572558.5 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC1700011171.hg.1 | ENST00000572558.5 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC1700011171.hg.1 | ENST00000572558.5 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC1700011171.hg.1 | ENST00000572558.5 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC1700011171.hg.1 | ENST00000572558.5 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC1700011171.hg.1 | ENST00000573912.1 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC1700011171.hg.1 | ENST00000573912.1 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC1700011171.hg.1 | ENST00000573912.1 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC1700011171.hg.1 | ENST00000573912.1 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC1700011171.hg.1 | ENST00000573912.1 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC1700011171.hg.1 | ENST00000574821.1 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC1700011171.hg.1 | ENST00000574821.1 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC1700011171.hg.1 | ENST00000574821.1 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC1700011171.hg.1 | ENST00000574821.1 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC1700011171.hg.1 | ENST00000574821.1 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC1700011171.hg.1 | ENST00000574989.1 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC1700011171.hg.1 | ENST00000576084.5 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC1700011171.hg.1 | ENST00000576084.5 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC1700011171.hg.1 | ENST00000576084.5 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC1700011171.hg.1 | ENST00000576084.5 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC1700011171.hg.1 | ENST00000576370.5 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC1700011171.hg.1 | ENST00000576370.5 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC1700011171.hg.1 | ENST00000576370.5 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC1700011171.hg.1 | ENST00000576370.5 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC1700011171.hg.1 | ENST00000576370.5 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC1700011171.hg.1 | ENST00000576370.5 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC1700011171.hg.1 | ENST00000576370.5 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC1700011171.hg.1 | ENST00000576370.5 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC1700011171.hg.1 | ENST00000576370.5 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC1700011171.hg.1 | ENST00000576370.5 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC1700011171.hg.1 | ENST00000576370.5 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC1700011171.hg.1 | NM_001162861.2 | Refseq |
| Affymetrix Human Transcriptome Array 2.0 | TC1700011171.hg.1 | NM_001162861.2 | Refseq |
| Affymetrix Human Transcriptome Array 2.0 | TC1700011171.hg.1 | NM_001162861.2 | Refseq |
| Affymetrix Human Transcriptome Array 2.0 | TC1700011171.hg.1 | NM_001162861.2 | Refseq |
| Affymetrix Human Transcriptome Array 2.0 | TC1700011171.hg.1 | NM_001162861.2 | Refseq |
| Affymetrix Human Transcriptome Array 2.0 | TC1700011171.hg.1 | NM_001162861.2 | Refseq |
| Affymetrix Human Transcriptome Array 2.0 | TC1700011171.hg.1 | NM_001162862.2 | Refseq |
| Affymetrix Human Transcriptome Array 2.0 | TC1700011171.hg.1 | NM_001162862.2 | Refseq |
| Affymetrix Human Transcriptome Array 2.0 | TC1700011171.hg.1 | NM_001162862.2 | Refseq |
| Affymetrix Human Transcriptome Array 2.0 | TC1700011171.hg.1 | NM_001162862.2 | Refseq |
| Affymetrix Human Transcriptome Array 2.0 | TC1700011171.hg.1 | NM_001162862.2 | Refseq |
| Affymetrix Human Transcriptome Array 2.0 | TC1700011171.hg.1 | NM_001162862.2 | Refseq |
| Affymetrix Human Transcriptome Array 2.0 | TC1700011171.hg.1 | NM_004375.4 | Refseq |
| Affymetrix Human Transcriptome Array 2.0 | TC1700011171.hg.1 | NM_004375.4 | Refseq |
| Affymetrix Human Transcriptome Array 2.0 | TC1700011171.hg.1 | NM_004375.4 | Refseq |
| Affymetrix Human Transcriptome Array 2.0 | TC1700011171.hg.1 | NM_004375.4 | Refseq |
| Affymetrix Human Transcriptome Array 2.0 | TC1700011171.hg.1 | NM_004375.4 | Refseq |
| Affymetrix Human Transcriptome Array 2.0 | TC1700011171.hg.1 | NM_004375.4 | Refseq |
| Affymetrix Human Transcriptome Array 2.0 | TC1700011171.hg.1 | NM_004375.4 | Refseq |
| Affymetrix Human Transcriptome Array 2.0 | TC1700011171.hg.1 | NM_004375.4 | Refseq |
| Affymetrix Human Transcriptome Array 2.0 | TC1700011171.hg.1 | NM_004375.4 | Refseq |
| Affymetrix Human Transcriptome Array 2.0 | TC1700011171.hg.1 | NR_027941.2 | Refseq |
| Affymetrix Human Transcriptome Array 2.0 | TC1700011171.hg.1 | NR_027941.2 | Refseq |
| Affymetrix Human Transcriptome Array 2.0 | TC1700011171.hg.1 | NR_027941.2 | Refseq |
| Affymetrix Human Transcriptome Array 2.0 | TC1700011171.hg.1 | NR_027941.2 | Refseq |
| Affymetrix Human Transcriptome Array 2.0 | TC1700011171.hg.1 | NR_027941.2 | Refseq |
| Affymetrix Human Transcriptome Array 2.0 | TC1700011171.hg.1 | NR_027941.2 | Refseq |
| Affymetrix Human Transcriptome Array 2.0 | TC1700011171.hg.1 | NR_027941.2 | Refseq |
| Affymetrix Human Transcriptome Array 2.0 | TC1700011171.hg.1 | NR_027941.2 | Refseq |
| Affymetrix Human Transcriptome Array 2.0 | TC1700011171.hg.1 | NR_027941.2 | Refseq |
| Affymetrix Human Transcriptome Array 2.0 | TC1700011171.hg.1 | NR_027942.2 | Refseq |
| Affymetrix Human Transcriptome Array 2.0 | TC1700011171.hg.1 | NR_027942.2 | Refseq |
| Affymetrix Human Transcriptome Array 2.0 | TC1700011171.hg.1 | NR_027942.2 | Refseq |
| Affymetrix Human Transcriptome Array 2.0 | TC1700011171.hg.1 | NR_027942.2 | Refseq |
| Affymetrix Human Transcriptome Array 2.0 | TC1700011171.hg.1 | NR_027942.2 | Refseq |
| Affymetrix Human Transcriptome Array 2.0 | TC1700011171.hg.1 | NR_027942.2 | Refseq |
| Affymetrix Human Transcriptome Array 2.0 | TC1700011171.hg.1 | NR_027942.2 | Refseq |
| Affymetrix Human Transcriptome Array 2.0 | TC1700011171.hg.1 | NR_027942.2 | Refseq |
| Affymetrix Human Transcriptome Array 2.0 | TC1700011171.hg.1 | NR_027942.2 | Refseq |
| Affymetrix Human Clariom D Assay | TC1700011171.hg.1 | ENST00000299335.7 | Ensembl |
| Affymetrix Human Clariom D Assay | TC1700011171.hg.1 | ENST00000299335.7 | Ensembl |
| Affymetrix Human Clariom D Assay | TC1700011171.hg.1 | ENST00000299335.7 | Ensembl |
| Affymetrix Human Clariom D Assay | TC1700011171.hg.1 | ENST00000299335.7 | Ensembl |
| Affymetrix Human Clariom D Assay | TC1700011171.hg.1 | ENST00000299335.7 | Ensembl |
| Affymetrix Human Clariom D Assay | TC1700011171.hg.1 | ENST00000299335.7 | Ensembl |
| Affymetrix Human Clariom D Assay | TC1700011171.hg.1 | ENST00000299335.7 | Ensembl |
| Affymetrix Human Clariom D Assay | TC1700011171.hg.1 | ENST00000299335.7 | Ensembl |
| Affymetrix Human Clariom D Assay | TC1700011171.hg.1 | ENST00000299335.7 | Ensembl |
| Affymetrix Human Clariom D Assay | TC1700011171.hg.1 | ENST00000299335.7 | Ensembl |
| Affymetrix Human Clariom D Assay | TC1700011171.hg.1 | ENST00000571584.1 | Ensembl |
| Affymetrix Human Clariom D Assay | TC1700011171.hg.1 | ENST00000571584.1 | Ensembl |
| Affymetrix Human Clariom D Assay | TC1700011171.hg.1 | ENST00000571584.1 | Ensembl |
| Affymetrix Human Clariom D Assay | TC1700011171.hg.1 | ENST00000571584.1 | Ensembl |
| Affymetrix Human Clariom D Assay | TC1700011171.hg.1 | ENST00000571584.1 | Ensembl |
| Affymetrix Human Clariom D Assay | TC1700011171.hg.1 | ENST00000572088.1 | Ensembl |
| Affymetrix Human Clariom D Assay | TC1700011171.hg.1 | ENST00000572088.1 | Ensembl |
| Affymetrix Human Clariom D Assay | TC1700011171.hg.1 | ENST00000572088.1 | Ensembl |
| Affymetrix Human Clariom D Assay | TC1700011171.hg.1 | ENST00000572088.1 | Ensembl |
| Affymetrix Human Clariom D Assay | TC1700011171.hg.1 | ENST00000572088.1 | Ensembl |
| Affymetrix Human Clariom D Assay | TC1700011171.hg.1 | ENST00000572088.1 | Ensembl |
| Affymetrix Human Clariom D Assay | TC1700011171.hg.1 | ENST00000572558.5 | Ensembl |
| Affymetrix Human Clariom D Assay | TC1700011171.hg.1 | ENST00000572558.5 | Ensembl |
| Affymetrix Human Clariom D Assay | TC1700011171.hg.1 | ENST00000572558.5 | Ensembl |
| Affymetrix Human Clariom D Assay | TC1700011171.hg.1 | ENST00000572558.5 | Ensembl |
| Affymetrix Human Clariom D Assay | TC1700011171.hg.1 | ENST00000572558.5 | Ensembl |
| Affymetrix Human Clariom D Assay | TC1700011171.hg.1 | ENST00000572558.5 | Ensembl |
| Affymetrix Human Clariom D Assay | TC1700011171.hg.1 | ENST00000572558.5 | Ensembl |
| Affymetrix Human Clariom D Assay | TC1700011171.hg.1 | ENST00000572558.5 | Ensembl |
| Affymetrix Human Clariom D Assay | TC1700011171.hg.1 | ENST00000573912.1 | Ensembl |
| Affymetrix Human Clariom D Assay | TC1700011171.hg.1 | ENST00000573912.1 | Ensembl |
| Affymetrix Human Clariom D Assay | TC1700011171.hg.1 | ENST00000573912.1 | Ensembl |
| Affymetrix Human Clariom D Assay | TC1700011171.hg.1 | ENST00000573912.1 | Ensembl |
| Affymetrix Human Clariom D Assay | TC1700011171.hg.1 | ENST00000573912.1 | Ensembl |
| Affymetrix Human Clariom D Assay | TC1700011171.hg.1 | ENST00000574821.1 | Ensembl |
| Affymetrix Human Clariom D Assay | TC1700011171.hg.1 | ENST00000574821.1 | Ensembl |
| Affymetrix Human Clariom D Assay | TC1700011171.hg.1 | ENST00000574821.1 | Ensembl |
| Affymetrix Human Clariom D Assay | TC1700011171.hg.1 | ENST00000574821.1 | Ensembl |
| Affymetrix Human Clariom D Assay | TC1700011171.hg.1 | ENST00000574821.1 | Ensembl |
| Affymetrix Human Clariom D Assay | TC1700011171.hg.1 | ENST00000574989.1 | Ensembl |
| Affymetrix Human Clariom D Assay | TC1700011171.hg.1 | ENST00000576084.5 | Ensembl |
| Affymetrix Human Clariom D Assay | TC1700011171.hg.1 | ENST00000576084.5 | Ensembl |
| Affymetrix Human Clariom D Assay | TC1700011171.hg.1 | ENST00000576084.5 | Ensembl |
| Affymetrix Human Clariom D Assay | TC1700011171.hg.1 | ENST00000576084.5 | Ensembl |
| Affymetrix Human Clariom D Assay | TC1700011171.hg.1 | ENST00000576370.5 | Ensembl |
| Affymetrix Human Clariom D Assay | TC1700011171.hg.1 | ENST00000576370.5 | Ensembl |
| Affymetrix Human Clariom D Assay | TC1700011171.hg.1 | ENST00000576370.5 | Ensembl |
| Affymetrix Human Clariom D Assay | TC1700011171.hg.1 | ENST00000576370.5 | Ensembl |
| Affymetrix Human Clariom D Assay | TC1700011171.hg.1 | ENST00000576370.5 | Ensembl |
| Affymetrix Human Clariom D Assay | TC1700011171.hg.1 | ENST00000576370.5 | Ensembl |
| Affymetrix Human Clariom D Assay | TC1700011171.hg.1 | ENST00000576370.5 | Ensembl |
| Affymetrix Human Clariom D Assay | TC1700011171.hg.1 | ENST00000576370.5 | Ensembl |
| Affymetrix Human Clariom D Assay | TC1700011171.hg.1 | ENST00000576370.5 | Ensembl |
| Affymetrix Human Clariom D Assay | TC1700011171.hg.1 | ENST00000576370.5 | Ensembl |
| Affymetrix Human Clariom D Assay | TC1700011171.hg.1 | ENST00000576370.5 | Ensembl |
| Affymetrix Human Clariom D Assay | TC1700011171.hg.1 | NM_001162861.2 | Refseq |
| Affymetrix Human Clariom D Assay | TC1700011171.hg.1 | NM_001162861.2 | Refseq |
| Affymetrix Human Clariom D Assay | TC1700011171.hg.1 | NM_001162861.2 | Refseq |
| Affymetrix Human Clariom D Assay | TC1700011171.hg.1 | NM_001162861.2 | Refseq |
| Affymetrix Human Clariom D Assay | TC1700011171.hg.1 | NM_001162861.2 | Refseq |
| Affymetrix Human Clariom D Assay | TC1700011171.hg.1 | NM_001162861.2 | Refseq |
| Affymetrix Human Clariom D Assay | TC1700011171.hg.1 | NM_001162862.2 | Refseq |
| Affymetrix Human Clariom D Assay | TC1700011171.hg.1 | NM_001162862.2 | Refseq |
| Affymetrix Human Clariom D Assay | TC1700011171.hg.1 | NM_001162862.2 | Refseq |
| Affymetrix Human Clariom D Assay | TC1700011171.hg.1 | NM_001162862.2 | Refseq |
| Affymetrix Human Clariom D Assay | TC1700011171.hg.1 | NM_001162862.2 | Refseq |
| Affymetrix Human Clariom D Assay | TC1700011171.hg.1 | NM_001162862.2 | Refseq |
| Affymetrix Human Clariom D Assay | TC1700011171.hg.1 | NM_004375.4 | Refseq |
| Affymetrix Human Clariom D Assay | TC1700011171.hg.1 | NM_004375.4 | Refseq |
| Affymetrix Human Clariom D Assay | TC1700011171.hg.1 | NM_004375.4 | Refseq |
| Affymetrix Human Clariom D Assay | TC1700011171.hg.1 | NM_004375.4 | Refseq |
| Affymetrix Human Clariom D Assay | TC1700011171.hg.1 | NM_004375.4 | Refseq |
| Affymetrix Human Clariom D Assay | TC1700011171.hg.1 | NM_004375.4 | Refseq |
| Affymetrix Human Clariom D Assay | TC1700011171.hg.1 | NM_004375.4 | Refseq |
| Affymetrix Human Clariom D Assay | TC1700011171.hg.1 | NM_004375.4 | Refseq |
| Affymetrix Human Clariom D Assay | TC1700011171.hg.1 | NM_004375.4 | Refseq |
| Affymetrix Human Clariom D Assay | TC1700011171.hg.1 | NR_027941.2 | Refseq |
| Affymetrix Human Clariom D Assay | TC1700011171.hg.1 | NR_027941.2 | Refseq |
| Affymetrix Human Clariom D Assay | TC1700011171.hg.1 | NR_027941.2 | Refseq |
| Affymetrix Human Clariom D Assay | TC1700011171.hg.1 | NR_027941.2 | Refseq |
| Affymetrix Human Clariom D Assay | TC1700011171.hg.1 | NR_027941.2 | Refseq |
| Affymetrix Human Clariom D Assay | TC1700011171.hg.1 | NR_027941.2 | Refseq |
| Affymetrix Human Clariom D Assay | TC1700011171.hg.1 | NR_027941.2 | Refseq |
| Affymetrix Human Clariom D Assay | TC1700011171.hg.1 | NR_027941.2 | Refseq |
| Affymetrix Human Clariom D Assay | TC1700011171.hg.1 | NR_027941.2 | Refseq |
| Affymetrix Human Clariom D Assay | TC1700011171.hg.1 | NR_027942.2 | Refseq |
| Affymetrix Human Clariom D Assay | TC1700011171.hg.1 | NR_027942.2 | Refseq |
| Affymetrix Human Clariom D Assay | TC1700011171.hg.1 | NR_027942.2 | Refseq |
| Affymetrix Human Clariom D Assay | TC1700011171.hg.1 | NR_027942.2 | Refseq |
| Affymetrix Human Clariom D Assay | TC1700011171.hg.1 | NR_027942.2 | Refseq |
| Affymetrix Human Clariom D Assay | TC1700011171.hg.1 | NR_027942.2 | Refseq |
| Affymetrix Human Clariom D Assay | TC1700011171.hg.1 | NR_027942.2 | Refseq |
| Affymetrix Human Clariom D Assay | TC1700011171.hg.1 | NR_027942.2 | Refseq |
| Affymetrix Human Clariom D Assay | TC1700011171.hg.1 | NR_027942.2 | Refseq |

COX11
Genomic Map