Assay Details
| Gene Symbol: SMARCA5 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | NM_003601.3 | NP_003592.3 | 22 - 23 | 3447 | 138 | |
| GenBank mRNA | AB010882.1 | - | 22 - 23 | 3187 | 138 | |
| AF086544.1 | - | 1 - 2 | 175 | 138 | ||
| AK302841.1 | - | 23 - 24 | 3082 | 138 | ||
| AK316444.1 | - | 23 - 24 | 3249 | 138 | ||
| BC023144.1 | - | 22 - 23 | 3198 | 4340921 | 138 | |
More Information
Updated Mapping Information
This assay targets the same location as HTA transcript cluster(s) TC04000701.hg.1
Set Membership: |
|
Matching Ambion siRNAs:
Panther Classification:
Gene Ontology Categories:
Function(s)
Process(es)
chromatin silencing at rDNA
double-strand break repair
nucleosome assembly
chromatin remodeling
DNA-templated transcription, initiation
regulation of transcription from RNA polymerase II promoter
covalent chromatin modification
nucleosome positioning
CENP-A containing nucleosome assembly
ATP-dependent chromatin remodeling
positive regulation of gene expression, epigenetic
positive regulation of transcription, DNA-templated
DNA binding
helicase activity
protein binding
ATP binding
ATPase activity
nucleosome binding
histone binding
chromatin silencing at rDNA
double-strand break repair
nucleosome assembly
chromatin remodeling
DNA-templated transcription, initiation
regulation of transcription from RNA polymerase II promoter
covalent chromatin modification
nucleosome positioning
CENP-A containing nucleosome assembly
ATP-dependent chromatin remodeling
positive regulation of gene expression, epigenetic
positive regulation of transcription, DNA-templated
DNA binding
helicase activity
protein binding
ATP binding
ATPase activity
nucleosome binding
histone binding
MicroArray Probes:
Affymetrix
| Array Name | Probe IDs | Targets | Target Type |
|---|---|---|---|
| Affymetrix Human Transcriptome Array 2.0 | PSR04009983.hg.1 | TC04000701.hg.1 | HTA TC |
| Affymetrix Human Transcriptome Array 2.0 | PSR04009983.hg.1 | ENST00000283131.3 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | PSR04009983.hg.1 | ENST00000535006.1 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | PSR04009983.hg.1 | ENST00000536484.1 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | PSR04009983.hg.1 | NM_003601.3 | Refseq |
| Affymetrix Human Transcriptome Array 2.0 | TC0400008882.hg.1 | ENST00000283131.3 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0400008882.hg.1 | ENST00000283131.3 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0400008882.hg.1 | ENST00000283131.3 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0400008882.hg.1 | ENST00000283131.3 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0400008882.hg.1 | ENST00000283131.3 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0400008882.hg.1 | ENST00000283131.3 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0400008882.hg.1 | ENST00000283131.3 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0400008882.hg.1 | ENST00000283131.3 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0400008882.hg.1 | ENST00000283131.3 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0400008882.hg.1 | ENST00000283131.3 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0400008882.hg.1 | ENST00000283131.3 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0400008882.hg.1 | ENST00000283131.3 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0400008882.hg.1 | ENST00000283131.3 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0400008882.hg.1 | ENST00000283131.3 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0400008882.hg.1 | ENST00000283131.3 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0400008882.hg.1 | ENST00000283131.3 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0400008882.hg.1 | ENST00000283131.3 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0400008882.hg.1 | ENST00000283131.3 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0400008882.hg.1 | ENST00000283131.3 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0400008882.hg.1 | ENST00000283131.3 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0400008882.hg.1 | ENST00000283131.3 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0400008882.hg.1 | ENST00000283131.3 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0400008882.hg.1 | ENST00000283131.3 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0400008882.hg.1 | ENST00000283131.3 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0400008882.hg.1 | ENST00000283131.3 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0400008882.hg.1 | ENST00000283131.3 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0400008882.hg.1 | ENST00000283131.3 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0400008882.hg.1 | ENST00000283131.3 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0400008882.hg.1 | ENST00000283131.3 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0400008882.hg.1 | ENST00000283131.3 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0400008882.hg.1 | ENST00000508573.1 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0400008882.hg.1 | ENST00000508573.1 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0400008882.hg.1 | ENST00000508573.1 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0400008882.hg.1 | ENST00000515531.1 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0400008882.hg.1 | ENST00000515531.1 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0400008882.hg.1 | ENST00000515531.1 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0400008882.hg.1 | NM_003601.3 | Refseq |
| Affymetrix Human Transcriptome Array 2.0 | TC0400008882.hg.1 | NM_003601.3 | Refseq |
| Affymetrix Human Transcriptome Array 2.0 | TC0400008882.hg.1 | NM_003601.3 | Refseq |
| Affymetrix Human Transcriptome Array 2.0 | TC0400008882.hg.1 | NM_003601.3 | Refseq |
| Affymetrix Human Transcriptome Array 2.0 | TC0400008882.hg.1 | NM_003601.3 | Refseq |
| Affymetrix Human Transcriptome Array 2.0 | TC0400008882.hg.1 | NM_003601.3 | Refseq |
| Affymetrix Human Transcriptome Array 2.0 | TC0400008882.hg.1 | NM_003601.3 | Refseq |
| Affymetrix Human Transcriptome Array 2.0 | TC0400008882.hg.1 | NM_003601.3 | Refseq |
| Affymetrix Human Transcriptome Array 2.0 | TC0400008882.hg.1 | NM_003601.3 | Refseq |
| Affymetrix Human Transcriptome Array 2.0 | TC0400008882.hg.1 | NM_003601.3 | Refseq |
| Affymetrix Human Transcriptome Array 2.0 | TC0400008882.hg.1 | NM_003601.3 | Refseq |
| Affymetrix Human Transcriptome Array 2.0 | TC0400008882.hg.1 | NM_003601.3 | Refseq |
| Affymetrix Human Transcriptome Array 2.0 | TC0400008882.hg.1 | NM_003601.3 | Refseq |
| Affymetrix Human Transcriptome Array 2.0 | TC0400008882.hg.1 | NM_003601.3 | Refseq |
| Affymetrix Human Transcriptome Array 2.0 | TC0400008882.hg.1 | NM_003601.3 | Refseq |
| Affymetrix Human Transcriptome Array 2.0 | TC0400008882.hg.1 | NM_003601.3 | Refseq |
| Affymetrix Human Transcriptome Array 2.0 | TC0400008882.hg.1 | NM_003601.3 | Refseq |
| Affymetrix Human Transcriptome Array 2.0 | TC0400008882.hg.1 | NM_003601.3 | Refseq |
| Affymetrix Human Transcriptome Array 2.0 | TC0400008882.hg.1 | NM_003601.3 | Refseq |
| Affymetrix Human Transcriptome Array 2.0 | TC0400008882.hg.1 | NM_003601.3 | Refseq |
| Affymetrix Human Transcriptome Array 2.0 | TC0400008882.hg.1 | NM_003601.3 | Refseq |
| Affymetrix Human Transcriptome Array 2.0 | TC0400008882.hg.1 | NM_003601.3 | Refseq |
| Affymetrix Human Transcriptome Array 2.0 | TC0400008882.hg.1 | NM_003601.3 | Refseq |
| Affymetrix Human Transcriptome Array 2.0 | TC0400008882.hg.1 | NM_003601.3 | Refseq |
| Affymetrix Human Transcriptome Array 2.0 | TC0400008882.hg.1 | NM_003601.3 | Refseq |
| Affymetrix Human Transcriptome Array 2.0 | TC0400008882.hg.1 | NM_003601.3 | Refseq |
| Affymetrix Human Transcriptome Array 2.0 | TC0400008882.hg.1 | NM_003601.3 | Refseq |
| Affymetrix Human Transcriptome Array 2.0 | TC0400008882.hg.1 | NM_003601.3 | Refseq |
| Affymetrix Human Transcriptome Array 2.0 | TC0400008882.hg.1 | NM_003601.3 | Refseq |
| Affymetrix Human Transcriptome Array 2.0 | TC0400008882.hg.1 | NM_003601.3 | Refseq |
| Affymetrix Human Clariom D Assay | PSR0400160996.hg.1 | ENST00000283131.3 | Ensembl |
| Affymetrix Human Clariom D Assay | PSR0400160996.hg.1 | NM_003601.3 | Refseq |
| Affymetrix Human Clariom D Assay | PSR0400160996.hg.1 | ENST00000283131.3 | Ensembl |
| Affymetrix Human Clariom D Assay | PSR0400160996.hg.1 | NM_003601.3 | Refseq |
| Affymetrix Human Clariom D Assay | PSR0400160996.hg.1 | TC0400008882.hg.1 | Clariom D TC |
| Affymetrix Human Clariom D Assay | TC0400008882.hg.1 | ENST00000283131.3 | Ensembl |
| Affymetrix Human Clariom D Assay | TC0400008882.hg.1 | ENST00000283131.3 | Ensembl |
| Affymetrix Human Clariom D Assay | TC0400008882.hg.1 | ENST00000283131.3 | Ensembl |
| Affymetrix Human Clariom D Assay | TC0400008882.hg.1 | ENST00000283131.3 | Ensembl |
| Affymetrix Human Clariom D Assay | TC0400008882.hg.1 | ENST00000283131.3 | Ensembl |
| Affymetrix Human Clariom D Assay | TC0400008882.hg.1 | ENST00000283131.3 | Ensembl |
| Affymetrix Human Clariom D Assay | TC0400008882.hg.1 | ENST00000283131.3 | Ensembl |
| Affymetrix Human Clariom D Assay | TC0400008882.hg.1 | ENST00000283131.3 | Ensembl |
| Affymetrix Human Clariom D Assay | TC0400008882.hg.1 | ENST00000283131.3 | Ensembl |
| Affymetrix Human Clariom D Assay | TC0400008882.hg.1 | ENST00000283131.3 | Ensembl |
| Affymetrix Human Clariom D Assay | TC0400008882.hg.1 | ENST00000283131.3 | Ensembl |
| Affymetrix Human Clariom D Assay | TC0400008882.hg.1 | ENST00000283131.3 | Ensembl |
| Affymetrix Human Clariom D Assay | TC0400008882.hg.1 | ENST00000283131.3 | Ensembl |
| Affymetrix Human Clariom D Assay | TC0400008882.hg.1 | ENST00000283131.3 | Ensembl |
| Affymetrix Human Clariom D Assay | TC0400008882.hg.1 | ENST00000283131.3 | Ensembl |
| Affymetrix Human Clariom D Assay | TC0400008882.hg.1 | ENST00000283131.3 | Ensembl |
| Affymetrix Human Clariom D Assay | TC0400008882.hg.1 | ENST00000283131.3 | Ensembl |
| Affymetrix Human Clariom D Assay | TC0400008882.hg.1 | ENST00000283131.3 | Ensembl |
| Affymetrix Human Clariom D Assay | TC0400008882.hg.1 | ENST00000283131.3 | Ensembl |
| Affymetrix Human Clariom D Assay | TC0400008882.hg.1 | ENST00000283131.3 | Ensembl |
| Affymetrix Human Clariom D Assay | TC0400008882.hg.1 | ENST00000283131.3 | Ensembl |
| Affymetrix Human Clariom D Assay | TC0400008882.hg.1 | ENST00000283131.3 | Ensembl |
| Affymetrix Human Clariom D Assay | TC0400008882.hg.1 | ENST00000283131.3 | Ensembl |
| Affymetrix Human Clariom D Assay | TC0400008882.hg.1 | ENST00000283131.3 | Ensembl |
| Affymetrix Human Clariom D Assay | TC0400008882.hg.1 | ENST00000283131.3 | Ensembl |
| Affymetrix Human Clariom D Assay | TC0400008882.hg.1 | ENST00000283131.3 | Ensembl |
| Affymetrix Human Clariom D Assay | TC0400008882.hg.1 | ENST00000283131.3 | Ensembl |
| Affymetrix Human Clariom D Assay | TC0400008882.hg.1 | ENST00000283131.3 | Ensembl |
| Affymetrix Human Clariom D Assay | TC0400008882.hg.1 | ENST00000283131.3 | Ensembl |
| Affymetrix Human Clariom D Assay | TC0400008882.hg.1 | ENST00000283131.3 | Ensembl |
| Affymetrix Human Clariom D Assay | TC0400008882.hg.1 | ENST00000508573.1 | Ensembl |
| Affymetrix Human Clariom D Assay | TC0400008882.hg.1 | ENST00000508573.1 | Ensembl |
| Affymetrix Human Clariom D Assay | TC0400008882.hg.1 | ENST00000508573.1 | Ensembl |
| Affymetrix Human Clariom D Assay | TC0400008882.hg.1 | ENST00000515531.1 | Ensembl |
| Affymetrix Human Clariom D Assay | TC0400008882.hg.1 | ENST00000515531.1 | Ensembl |
| Affymetrix Human Clariom D Assay | TC0400008882.hg.1 | ENST00000515531.1 | Ensembl |
| Affymetrix Human Clariom D Assay | TC0400008882.hg.1 | NM_003601.3 | Refseq |
| Affymetrix Human Clariom D Assay | TC0400008882.hg.1 | NM_003601.3 | Refseq |
| Affymetrix Human Clariom D Assay | TC0400008882.hg.1 | NM_003601.3 | Refseq |
| Affymetrix Human Clariom D Assay | TC0400008882.hg.1 | NM_003601.3 | Refseq |
| Affymetrix Human Clariom D Assay | TC0400008882.hg.1 | NM_003601.3 | Refseq |
| Affymetrix Human Clariom D Assay | TC0400008882.hg.1 | NM_003601.3 | Refseq |
| Affymetrix Human Clariom D Assay | TC0400008882.hg.1 | NM_003601.3 | Refseq |
| Affymetrix Human Clariom D Assay | TC0400008882.hg.1 | NM_003601.3 | Refseq |
| Affymetrix Human Clariom D Assay | TC0400008882.hg.1 | NM_003601.3 | Refseq |
| Affymetrix Human Clariom D Assay | TC0400008882.hg.1 | NM_003601.3 | Refseq |
| Affymetrix Human Clariom D Assay | TC0400008882.hg.1 | NM_003601.3 | Refseq |
| Affymetrix Human Clariom D Assay | TC0400008882.hg.1 | NM_003601.3 | Refseq |
| Affymetrix Human Clariom D Assay | TC0400008882.hg.1 | NM_003601.3 | Refseq |
| Affymetrix Human Clariom D Assay | TC0400008882.hg.1 | NM_003601.3 | Refseq |
| Affymetrix Human Clariom D Assay | TC0400008882.hg.1 | NM_003601.3 | Refseq |
| Affymetrix Human Clariom D Assay | TC0400008882.hg.1 | NM_003601.3 | Refseq |
| Affymetrix Human Clariom D Assay | TC0400008882.hg.1 | NM_003601.3 | Refseq |
| Affymetrix Human Clariom D Assay | TC0400008882.hg.1 | NM_003601.3 | Refseq |
| Affymetrix Human Clariom D Assay | TC0400008882.hg.1 | NM_003601.3 | Refseq |
| Affymetrix Human Clariom D Assay | TC0400008882.hg.1 | NM_003601.3 | Refseq |
| Affymetrix Human Clariom D Assay | TC0400008882.hg.1 | NM_003601.3 | Refseq |
| Affymetrix Human Clariom D Assay | TC0400008882.hg.1 | NM_003601.3 | Refseq |
| Affymetrix Human Clariom D Assay | TC0400008882.hg.1 | NM_003601.3 | Refseq |
| Affymetrix Human Clariom D Assay | TC0400008882.hg.1 | NM_003601.3 | Refseq |
| Affymetrix Human Clariom D Assay | TC0400008882.hg.1 | NM_003601.3 | Refseq |
| Affymetrix Human Clariom D Assay | TC0400008882.hg.1 | NM_003601.3 | Refseq |
| Affymetrix Human Clariom D Assay | TC0400008882.hg.1 | NM_003601.3 | Refseq |
| Affymetrix Human Clariom D Assay | TC0400008882.hg.1 | NM_003601.3 | Refseq |
| Affymetrix Human Clariom D Assay | TC0400008882.hg.1 | NM_003601.3 | Refseq |
| Affymetrix Human Clariom D Assay | TC0400008882.hg.1 | NM_003601.3 | Refseq |

SMARCA5
Genomic Map