Assay Details
| Gene Symbol: ST3GAL6 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | NM_001271145.1 | NP_001258074.1 | 1 - 2 | 306 | 55 | |
| NM_001323363.1 | NP_001310292.1 | 1 - 2 | 306 | 55 | ||
| GenBank mRNA | AF119391.1 | - | 1 - 2 | 289 | 55 | |
More Information
Additional Information:
This assay is in the 5'UTR region of the AF119391.1, NM_001323363.1 transcripts.
Set Membership: |
|

ST3GAL6
Genomic Map