Assay Details
| Gene Symbol: IARS2 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | NM_018060.3 | NP_060530.3 | 14 - 15 | 1937 | 82 | |
| GenBank mRNA | AK001188.1 | - | 12 - 13 | 1412 | 82 | |
| AK001726.1 | - | 8 - 9 | 925 | 82 | ||
| AK022665.1 | - | 4 - 5 | 504 | 82 | ||
| AK291432.1 | - | 14 - 15 | 1746 | 82 | ||
| BC010218.1 | - | 10 - 11 | 1117 | 82 | ||
| BC040376.1 | - | 14 - 15 | 1670 | 82 | ||
| BC047880.1 | - | 13 - 14 | 1464 | 82 | ||
| BC137438.1 | - | 14 - 15 | 1907 | 9021436 | 82 | |
More Information
Updated Mapping Information
This assay targets the same location as HTA transcript cluster(s) TC01001814.hg.1
Set Membership: |
|
Matching Ambion siRNAs:
Panther Classification:
Gene Ontology Categories:
MicroArray Probes:
Affymetrix
| Array Name | Probe IDs | Targets | Target Type |
|---|---|---|---|
| Affymetrix Human Transcriptome Array 2.0 | PSR01028241.hg.1 | TC01001814.hg.1 | HTA TC |
| Affymetrix Human Transcriptome Array 2.0 | PSR01028241.hg.1 | ENST00000302637.5 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | PSR01028241.hg.1 | ENST00000366922.1 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | PSR01028241.hg.1 | NM_018060.3 | Refseq |
| Affymetrix Human Transcriptome Array 2.0 | TC0100011648.hg.1 | ENST00000366922.2 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0100011648.hg.1 | ENST00000366922.2 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0100011648.hg.1 | ENST00000366922.2 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0100011648.hg.1 | ENST00000366922.2 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0100011648.hg.1 | ENST00000366922.2 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0100011648.hg.1 | ENST00000366922.2 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0100011648.hg.1 | ENST00000366922.2 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0100011648.hg.1 | ENST00000366922.2 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0100011648.hg.1 | ENST00000366922.2 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0100011648.hg.1 | ENST00000366922.2 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0100011648.hg.1 | ENST00000366922.2 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0100011648.hg.1 | ENST00000366922.2 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0100011648.hg.1 | ENST00000366922.2 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0100011648.hg.1 | ENST00000366922.2 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0100011648.hg.1 | ENST00000366922.2 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0100011648.hg.1 | ENST00000366922.2 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0100011648.hg.1 | ENST00000366922.2 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0100011648.hg.1 | ENST00000366922.2 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0100011648.hg.1 | ENST00000366922.2 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0100011648.hg.1 | ENST00000366922.2 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0100011648.hg.1 | ENST00000366922.2 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0100011648.hg.1 | ENST00000366922.2 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0100011648.hg.1 | ENST00000366922.2 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0100011648.hg.1 | ENST00000366922.2 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0100011648.hg.1 | ENST00000366922.2 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0100011648.hg.1 | ENST00000366922.2 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0100011648.hg.1 | ENST00000366922.2 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0100011648.hg.1 | ENST00000366922.2 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0100011648.hg.1 | ENST00000467924.1 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0100011648.hg.1 | ENST00000467924.1 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0100011648.hg.1 | ENST00000467924.1 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0100011648.hg.1 | ENST00000467924.1 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0100011648.hg.1 | ENST00000467924.1 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0100011648.hg.1 | ENST00000488777.1 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0100011648.hg.1 | ENST00000488777.1 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0100011648.hg.1 | ENST00000488777.1 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0100011648.hg.1 | ENST00000488777.1 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0100011648.hg.1 | ENST00000490891.1 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0100011648.hg.1 | ENST00000490891.1 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0100011648.hg.1 | ENST00000490891.1 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0100011648.hg.1 | ENST00000490891.1 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0100011648.hg.1 | ENST00000490891.1 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0100011648.hg.1 | NM_018060.3 | Refseq |
| Affymetrix Human Transcriptome Array 2.0 | TC0100011648.hg.1 | NM_018060.3 | Refseq |
| Affymetrix Human Transcriptome Array 2.0 | TC0100011648.hg.1 | NM_018060.3 | Refseq |
| Affymetrix Human Transcriptome Array 2.0 | TC0100011648.hg.1 | NM_018060.3 | Refseq |
| Affymetrix Human Transcriptome Array 2.0 | TC0100011648.hg.1 | NM_018060.3 | Refseq |
| Affymetrix Human Transcriptome Array 2.0 | TC0100011648.hg.1 | NM_018060.3 | Refseq |
| Affymetrix Human Transcriptome Array 2.0 | TC0100011648.hg.1 | NM_018060.3 | Refseq |
| Affymetrix Human Transcriptome Array 2.0 | TC0100011648.hg.1 | NM_018060.3 | Refseq |
| Affymetrix Human Transcriptome Array 2.0 | TC0100011648.hg.1 | NM_018060.3 | Refseq |
| Affymetrix Human Transcriptome Array 2.0 | TC0100011648.hg.1 | NM_018060.3 | Refseq |
| Affymetrix Human Transcriptome Array 2.0 | TC0100011648.hg.1 | NM_018060.3 | Refseq |
| Affymetrix Human Transcriptome Array 2.0 | TC0100011648.hg.1 | NM_018060.3 | Refseq |
| Affymetrix Human Transcriptome Array 2.0 | TC0100011648.hg.1 | NM_018060.3 | Refseq |
| Affymetrix Human Transcriptome Array 2.0 | TC0100011648.hg.1 | NM_018060.3 | Refseq |
| Affymetrix Human Transcriptome Array 2.0 | TC0100011648.hg.1 | NM_018060.3 | Refseq |
| Affymetrix Human Transcriptome Array 2.0 | TC0100011648.hg.1 | NM_018060.3 | Refseq |
| Affymetrix Human Transcriptome Array 2.0 | TC0100011648.hg.1 | NM_018060.3 | Refseq |
| Affymetrix Human Transcriptome Array 2.0 | TC0100011648.hg.1 | NM_018060.3 | Refseq |
| Affymetrix Human Transcriptome Array 2.0 | TC0100011648.hg.1 | NM_018060.3 | Refseq |
| Affymetrix Human Transcriptome Array 2.0 | TC0100011648.hg.1 | NM_018060.3 | Refseq |
| Affymetrix Human Transcriptome Array 2.0 | TC0100011648.hg.1 | NM_018060.3 | Refseq |
| Affymetrix Human Transcriptome Array 2.0 | TC0100011648.hg.1 | NM_018060.3 | Refseq |
| Affymetrix Human Transcriptome Array 2.0 | TC0100011648.hg.1 | NM_018060.3 | Refseq |
| Affymetrix Human Transcriptome Array 2.0 | TC0100011648.hg.1 | NM_018060.3 | Refseq |
| Affymetrix Human Transcriptome Array 2.0 | TC0100011648.hg.1 | NM_018060.3 | Refseq |
| Affymetrix Human Transcriptome Array 2.0 | TC0100011648.hg.1 | NM_018060.3 | Refseq |
| Affymetrix Human Transcriptome Array 2.0 | TC0100011648.hg.1 | NM_018060.3 | Refseq |
| Affymetrix Human Transcriptome Array 2.0 | TC0100011648.hg.1 | NM_018060.3 | Refseq |
| Affymetrix Human Clariom D Assay | TC0100011648.hg.1 | ENST00000366922.2 | Ensembl |
| Affymetrix Human Clariom D Assay | TC0100011648.hg.1 | ENST00000366922.2 | Ensembl |
| Affymetrix Human Clariom D Assay | TC0100011648.hg.1 | ENST00000366922.2 | Ensembl |
| Affymetrix Human Clariom D Assay | TC0100011648.hg.1 | ENST00000366922.2 | Ensembl |
| Affymetrix Human Clariom D Assay | TC0100011648.hg.1 | ENST00000366922.2 | Ensembl |
| Affymetrix Human Clariom D Assay | TC0100011648.hg.1 | ENST00000366922.2 | Ensembl |
| Affymetrix Human Clariom D Assay | TC0100011648.hg.1 | ENST00000366922.2 | Ensembl |
| Affymetrix Human Clariom D Assay | TC0100011648.hg.1 | ENST00000366922.2 | Ensembl |
| Affymetrix Human Clariom D Assay | TC0100011648.hg.1 | ENST00000366922.2 | Ensembl |
| Affymetrix Human Clariom D Assay | TC0100011648.hg.1 | ENST00000366922.2 | Ensembl |
| Affymetrix Human Clariom D Assay | TC0100011648.hg.1 | ENST00000366922.2 | Ensembl |
| Affymetrix Human Clariom D Assay | TC0100011648.hg.1 | ENST00000366922.2 | Ensembl |
| Affymetrix Human Clariom D Assay | TC0100011648.hg.1 | ENST00000366922.2 | Ensembl |
| Affymetrix Human Clariom D Assay | TC0100011648.hg.1 | ENST00000366922.2 | Ensembl |
| Affymetrix Human Clariom D Assay | TC0100011648.hg.1 | ENST00000366922.2 | Ensembl |
| Affymetrix Human Clariom D Assay | TC0100011648.hg.1 | ENST00000366922.2 | Ensembl |
| Affymetrix Human Clariom D Assay | TC0100011648.hg.1 | ENST00000366922.2 | Ensembl |
| Affymetrix Human Clariom D Assay | TC0100011648.hg.1 | ENST00000366922.2 | Ensembl |
| Affymetrix Human Clariom D Assay | TC0100011648.hg.1 | ENST00000366922.2 | Ensembl |
| Affymetrix Human Clariom D Assay | TC0100011648.hg.1 | ENST00000366922.2 | Ensembl |
| Affymetrix Human Clariom D Assay | TC0100011648.hg.1 | ENST00000366922.2 | Ensembl |
| Affymetrix Human Clariom D Assay | TC0100011648.hg.1 | ENST00000366922.2 | Ensembl |
| Affymetrix Human Clariom D Assay | TC0100011648.hg.1 | ENST00000366922.2 | Ensembl |
| Affymetrix Human Clariom D Assay | TC0100011648.hg.1 | ENST00000366922.2 | Ensembl |
| Affymetrix Human Clariom D Assay | TC0100011648.hg.1 | ENST00000366922.2 | Ensembl |
| Affymetrix Human Clariom D Assay | TC0100011648.hg.1 | ENST00000366922.2 | Ensembl |
| Affymetrix Human Clariom D Assay | TC0100011648.hg.1 | ENST00000366922.2 | Ensembl |
| Affymetrix Human Clariom D Assay | TC0100011648.hg.1 | ENST00000366922.2 | Ensembl |
| Affymetrix Human Clariom D Assay | TC0100011648.hg.1 | ENST00000467924.1 | Ensembl |
| Affymetrix Human Clariom D Assay | TC0100011648.hg.1 | ENST00000467924.1 | Ensembl |
| Affymetrix Human Clariom D Assay | TC0100011648.hg.1 | ENST00000467924.1 | Ensembl |
| Affymetrix Human Clariom D Assay | TC0100011648.hg.1 | ENST00000467924.1 | Ensembl |
| Affymetrix Human Clariom D Assay | TC0100011648.hg.1 | ENST00000467924.1 | Ensembl |
| Affymetrix Human Clariom D Assay | TC0100011648.hg.1 | ENST00000488777.1 | Ensembl |
| Affymetrix Human Clariom D Assay | TC0100011648.hg.1 | ENST00000488777.1 | Ensembl |
| Affymetrix Human Clariom D Assay | TC0100011648.hg.1 | ENST00000488777.1 | Ensembl |
| Affymetrix Human Clariom D Assay | TC0100011648.hg.1 | ENST00000488777.1 | Ensembl |
| Affymetrix Human Clariom D Assay | TC0100011648.hg.1 | ENST00000490891.1 | Ensembl |
| Affymetrix Human Clariom D Assay | TC0100011648.hg.1 | ENST00000490891.1 | Ensembl |
| Affymetrix Human Clariom D Assay | TC0100011648.hg.1 | ENST00000490891.1 | Ensembl |
| Affymetrix Human Clariom D Assay | TC0100011648.hg.1 | ENST00000490891.1 | Ensembl |
| Affymetrix Human Clariom D Assay | TC0100011648.hg.1 | ENST00000490891.1 | Ensembl |
| Affymetrix Human Clariom D Assay | TC0100011648.hg.1 | NM_018060.3 | Refseq |
| Affymetrix Human Clariom D Assay | TC0100011648.hg.1 | NM_018060.3 | Refseq |
| Affymetrix Human Clariom D Assay | TC0100011648.hg.1 | NM_018060.3 | Refseq |
| Affymetrix Human Clariom D Assay | TC0100011648.hg.1 | NM_018060.3 | Refseq |
| Affymetrix Human Clariom D Assay | TC0100011648.hg.1 | NM_018060.3 | Refseq |
| Affymetrix Human Clariom D Assay | TC0100011648.hg.1 | NM_018060.3 | Refseq |
| Affymetrix Human Clariom D Assay | TC0100011648.hg.1 | NM_018060.3 | Refseq |
| Affymetrix Human Clariom D Assay | TC0100011648.hg.1 | NM_018060.3 | Refseq |
| Affymetrix Human Clariom D Assay | TC0100011648.hg.1 | NM_018060.3 | Refseq |
| Affymetrix Human Clariom D Assay | TC0100011648.hg.1 | NM_018060.3 | Refseq |
| Affymetrix Human Clariom D Assay | TC0100011648.hg.1 | NM_018060.3 | Refseq |
| Affymetrix Human Clariom D Assay | TC0100011648.hg.1 | NM_018060.3 | Refseq |
| Affymetrix Human Clariom D Assay | TC0100011648.hg.1 | NM_018060.3 | Refseq |
| Affymetrix Human Clariom D Assay | TC0100011648.hg.1 | NM_018060.3 | Refseq |
| Affymetrix Human Clariom D Assay | TC0100011648.hg.1 | NM_018060.3 | Refseq |
| Affymetrix Human Clariom D Assay | TC0100011648.hg.1 | NM_018060.3 | Refseq |
| Affymetrix Human Clariom D Assay | TC0100011648.hg.1 | NM_018060.3 | Refseq |
| Affymetrix Human Clariom D Assay | TC0100011648.hg.1 | NM_018060.3 | Refseq |
| Affymetrix Human Clariom D Assay | TC0100011648.hg.1 | NM_018060.3 | Refseq |
| Affymetrix Human Clariom D Assay | TC0100011648.hg.1 | NM_018060.3 | Refseq |
| Affymetrix Human Clariom D Assay | TC0100011648.hg.1 | NM_018060.3 | Refseq |
| Affymetrix Human Clariom D Assay | TC0100011648.hg.1 | NM_018060.3 | Refseq |
| Affymetrix Human Clariom D Assay | TC0100011648.hg.1 | NM_018060.3 | Refseq |
| Affymetrix Human Clariom D Assay | TC0100011648.hg.1 | NM_018060.3 | Refseq |
| Affymetrix Human Clariom D Assay | TC0100011648.hg.1 | NM_018060.3 | Refseq |
| Affymetrix Human Clariom D Assay | TC0100011648.hg.1 | NM_018060.3 | Refseq |
| Affymetrix Human Clariom D Assay | TC0100011648.hg.1 | NM_018060.3 | Refseq |
| Affymetrix Human Clariom D Assay | TC0100011648.hg.1 | NM_018060.3 | Refseq |
| Affymetrix Human Clariom D Assay | PSR0100185746.hg.1 | ENST00000366922.2 | Ensembl |
| Affymetrix Human Clariom D Assay | PSR0100185746.hg.1 | ENST00000490891.1 | Ensembl |
| Affymetrix Human Clariom D Assay | PSR0100185746.hg.1 | NM_018060.3 | Refseq |
| Affymetrix Human Clariom D Assay | PSR0100185746.hg.1 | ENST00000366922.2 | Ensembl |
| Affymetrix Human Clariom D Assay | PSR0100185746.hg.1 | ENST00000490891.1 | Ensembl |
| Affymetrix Human Clariom D Assay | PSR0100185746.hg.1 | NM_018060.3 | Refseq |
| Affymetrix Human Clariom D Assay | PSR0100185746.hg.1 | TC0100011648.hg.1 | Clariom D TC |

IARS2
Genomic Map