Assay Details
| Gene Symbol: PRG2 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| GenBank mRNA | Z26248.1 | - | 7 - 8 | 772 | 70 | |
More Information
Additional Information:
This assay is in the 5'UTR region of the Z26248.1 transcripts.
Set Membership: |
|

PRG2
Genomic Map