Assay Details
| Gene Symbol: CNTFR | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | NM_001207011.1 | NP_001193940.1 | 1 - 2 | 57 | 109 | |
| XM_017014262.1 | XP_016869751.1 | 1 - 2 | 70 | 109 | ||
More Information
Updated Mapping Information
This assay targets the same location as HTA transcript cluster(s) TC09001038.hg
Additional Information:
This assay is in the 5'UTR region of the NM_001207011.1,XM_017014262.1 transcripts.
Set Membership: |
|
Panther Classification:
Gene Ontology Categories:
Function(s)
Process(es)
suckling behavior
brainstem development
signal transduction
nervous system development
sex differentiation
positive regulation of cell proliferation
negative regulation of neuron apoptotic process
skeletal muscle organ development
ciliary neurotrophic factor-mediated signaling pathway
ciliary neurotrophic factor receptor activity
receptor binding
protein binding
cytokine binding
suckling behavior
brainstem development
signal transduction
nervous system development
sex differentiation
positive regulation of cell proliferation
negative regulation of neuron apoptotic process
skeletal muscle organ development
ciliary neurotrophic factor-mediated signaling pathway
ciliary neurotrophic factor receptor activity
receptor binding
protein binding
cytokine binding
MicroArray Probes:
Affymetrix
| Array Name | Probe IDs | Targets | Target Type |
|---|---|---|---|
| Affymetrix Human Transcriptome Array 2.0 | PSR09013610.hg.1 | TC09001038.hg.1 | HTA TC |
| Affymetrix Human Transcriptome Array 2.0 | PSR09013610.hg.1 | NM_001207011.1 | Refseq |
| Affymetrix Human Transcriptome Array 2.0 | TC0900009905.hg.1 | ENST00000351266.8 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0900009905.hg.1 | ENST00000351266.8 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0900009905.hg.1 | ENST00000351266.8 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0900009905.hg.1 | ENST00000351266.8 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0900009905.hg.1 | ENST00000351266.8 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0900009905.hg.1 | ENST00000351266.8 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0900009905.hg.1 | ENST00000351266.8 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0900009905.hg.1 | ENST00000351266.8 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0900009905.hg.1 | ENST00000351266.8 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0900009905.hg.1 | ENST00000378980.7 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0900009905.hg.1 | ENST00000378980.7 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0900009905.hg.1 | ENST00000378980.7 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0900009905.hg.1 | ENST00000378980.7 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0900009905.hg.1 | ENST00000378980.7 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0900009905.hg.1 | ENST00000378980.7 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0900009905.hg.1 | ENST00000378980.7 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0900009905.hg.1 | ENST00000378980.7 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0900009905.hg.1 | ENST00000378980.7 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0900009905.hg.1 | ENST00000378980.7 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0900009905.hg.1 | ENST00000378980.7 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0900009905.hg.1 | ENST00000417345.2 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0900009905.hg.1 | ENST00000417345.2 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0900009905.hg.1 | ENST00000417345.2 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0900009905.hg.1 | ENST00000417345.2 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0900009905.hg.1 | ENST00000417345.2 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0900009905.hg.1 | ENST00000417345.2 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0900009905.hg.1 | ENST00000417345.2 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0900009905.hg.1 | ENST00000610543.4 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0900009905.hg.1 | ENST00000610543.4 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0900009905.hg.1 | ENST00000610543.4 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0900009905.hg.1 | ENST00000610543.4 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0900009905.hg.1 | ENST00000610543.4 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0900009905.hg.1 | ENST00000610543.4 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0900009905.hg.1 | ENST00000610543.4 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0900009905.hg.1 | ENST00000610543.4 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0900009905.hg.1 | ENST00000610543.4 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0900009905.hg.1 | ENST00000610543.4 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0900009905.hg.1 | NM_001207011.1 | Refseq |
| Affymetrix Human Transcriptome Array 2.0 | TC0900009905.hg.1 | NM_001207011.1 | Refseq |
| Affymetrix Human Transcriptome Array 2.0 | TC0900009905.hg.1 | NM_001207011.1 | Refseq |
| Affymetrix Human Transcriptome Array 2.0 | TC0900009905.hg.1 | NM_001207011.1 | Refseq |
| Affymetrix Human Transcriptome Array 2.0 | TC0900009905.hg.1 | NM_001207011.1 | Refseq |
| Affymetrix Human Transcriptome Array 2.0 | TC0900009905.hg.1 | NM_001207011.1 | Refseq |
| Affymetrix Human Transcriptome Array 2.0 | TC0900009905.hg.1 | NM_001207011.1 | Refseq |
| Affymetrix Human Transcriptome Array 2.0 | TC0900009905.hg.1 | NM_001207011.1 | Refseq |
| Affymetrix Human Transcriptome Array 2.0 | TC0900009905.hg.1 | NM_001207011.1 | Refseq |
| Affymetrix Human Transcriptome Array 2.0 | TC0900009905.hg.1 | NM_001207011.1 | Refseq |
| Affymetrix Human Transcriptome Array 2.0 | TC0900009905.hg.1 | NM_001842.4 | Refseq |
| Affymetrix Human Transcriptome Array 2.0 | TC0900009905.hg.1 | NM_001842.4 | Refseq |
| Affymetrix Human Transcriptome Array 2.0 | TC0900009905.hg.1 | NM_001842.4 | Refseq |
| Affymetrix Human Transcriptome Array 2.0 | TC0900009905.hg.1 | NM_001842.4 | Refseq |
| Affymetrix Human Transcriptome Array 2.0 | TC0900009905.hg.1 | NM_001842.4 | Refseq |
| Affymetrix Human Transcriptome Array 2.0 | TC0900009905.hg.1 | NM_001842.4 | Refseq |
| Affymetrix Human Transcriptome Array 2.0 | TC0900009905.hg.1 | NM_001842.4 | Refseq |
| Affymetrix Human Transcriptome Array 2.0 | TC0900009905.hg.1 | NM_001842.4 | Refseq |
| Affymetrix Human Transcriptome Array 2.0 | TC0900009905.hg.1 | NM_001842.4 | Refseq |
| Affymetrix Human Transcriptome Array 2.0 | TC0900009905.hg.1 | NM_001842.4 | Refseq |
| Affymetrix Human Transcriptome Array 2.0 | TC0900009905.hg.1 | NM_147164.2 | Refseq |
| Affymetrix Human Transcriptome Array 2.0 | TC0900009905.hg.1 | NM_147164.2 | Refseq |
| Affymetrix Human Transcriptome Array 2.0 | TC0900009905.hg.1 | NM_147164.2 | Refseq |
| Affymetrix Human Transcriptome Array 2.0 | TC0900009905.hg.1 | NM_147164.2 | Refseq |
| Affymetrix Human Transcriptome Array 2.0 | TC0900009905.hg.1 | NM_147164.2 | Refseq |
| Affymetrix Human Transcriptome Array 2.0 | TC0900009905.hg.1 | NM_147164.2 | Refseq |
| Affymetrix Human Transcriptome Array 2.0 | TC0900009905.hg.1 | NM_147164.2 | Refseq |
| Affymetrix Human Transcriptome Array 2.0 | TC0900009905.hg.1 | NM_147164.2 | Refseq |
| Affymetrix Human Transcriptome Array 2.0 | TC0900009905.hg.1 | NM_147164.2 | Refseq |
| Affymetrix Human Transcriptome Array 2.0 | TC0900009905.hg.1 | NM_147164.2 | Refseq |
| Affymetrix Human Transcriptome Array 2.0 | TC0900009905.hg.1 | NM_147164.2 | Refseq |
| Affymetrix Human Clariom D Assay | TC0900009905.hg.1 | ENST00000351266.8 | Ensembl |
| Affymetrix Human Clariom D Assay | TC0900009905.hg.1 | ENST00000351266.8 | Ensembl |
| Affymetrix Human Clariom D Assay | TC0900009905.hg.1 | ENST00000351266.8 | Ensembl |
| Affymetrix Human Clariom D Assay | TC0900009905.hg.1 | ENST00000351266.8 | Ensembl |
| Affymetrix Human Clariom D Assay | TC0900009905.hg.1 | ENST00000351266.8 | Ensembl |
| Affymetrix Human Clariom D Assay | TC0900009905.hg.1 | ENST00000351266.8 | Ensembl |
| Affymetrix Human Clariom D Assay | TC0900009905.hg.1 | ENST00000351266.8 | Ensembl |
| Affymetrix Human Clariom D Assay | TC0900009905.hg.1 | ENST00000351266.8 | Ensembl |
| Affymetrix Human Clariom D Assay | TC0900009905.hg.1 | ENST00000351266.8 | Ensembl |
| Affymetrix Human Clariom D Assay | TC0900009905.hg.1 | ENST00000378980.7 | Ensembl |
| Affymetrix Human Clariom D Assay | TC0900009905.hg.1 | ENST00000378980.7 | Ensembl |
| Affymetrix Human Clariom D Assay | TC0900009905.hg.1 | ENST00000378980.7 | Ensembl |
| Affymetrix Human Clariom D Assay | TC0900009905.hg.1 | ENST00000378980.7 | Ensembl |
| Affymetrix Human Clariom D Assay | TC0900009905.hg.1 | ENST00000378980.7 | Ensembl |
| Affymetrix Human Clariom D Assay | TC0900009905.hg.1 | ENST00000378980.7 | Ensembl |
| Affymetrix Human Clariom D Assay | TC0900009905.hg.1 | ENST00000378980.7 | Ensembl |
| Affymetrix Human Clariom D Assay | TC0900009905.hg.1 | ENST00000378980.7 | Ensembl |
| Affymetrix Human Clariom D Assay | TC0900009905.hg.1 | ENST00000378980.7 | Ensembl |
| Affymetrix Human Clariom D Assay | TC0900009905.hg.1 | ENST00000378980.7 | Ensembl |
| Affymetrix Human Clariom D Assay | TC0900009905.hg.1 | ENST00000378980.7 | Ensembl |
| Affymetrix Human Clariom D Assay | TC0900009905.hg.1 | ENST00000417345.2 | Ensembl |
| Affymetrix Human Clariom D Assay | TC0900009905.hg.1 | ENST00000417345.2 | Ensembl |
| Affymetrix Human Clariom D Assay | TC0900009905.hg.1 | ENST00000417345.2 | Ensembl |
| Affymetrix Human Clariom D Assay | TC0900009905.hg.1 | ENST00000417345.2 | Ensembl |
| Affymetrix Human Clariom D Assay | TC0900009905.hg.1 | ENST00000417345.2 | Ensembl |
| Affymetrix Human Clariom D Assay | TC0900009905.hg.1 | ENST00000417345.2 | Ensembl |
| Affymetrix Human Clariom D Assay | TC0900009905.hg.1 | ENST00000417345.2 | Ensembl |
| Affymetrix Human Clariom D Assay | TC0900009905.hg.1 | ENST00000610543.4 | Ensembl |
| Affymetrix Human Clariom D Assay | TC0900009905.hg.1 | ENST00000610543.4 | Ensembl |
| Affymetrix Human Clariom D Assay | TC0900009905.hg.1 | ENST00000610543.4 | Ensembl |
| Affymetrix Human Clariom D Assay | TC0900009905.hg.1 | ENST00000610543.4 | Ensembl |
| Affymetrix Human Clariom D Assay | TC0900009905.hg.1 | ENST00000610543.4 | Ensembl |
| Affymetrix Human Clariom D Assay | TC0900009905.hg.1 | ENST00000610543.4 | Ensembl |
| Affymetrix Human Clariom D Assay | TC0900009905.hg.1 | ENST00000610543.4 | Ensembl |
| Affymetrix Human Clariom D Assay | TC0900009905.hg.1 | ENST00000610543.4 | Ensembl |
| Affymetrix Human Clariom D Assay | TC0900009905.hg.1 | ENST00000610543.4 | Ensembl |
| Affymetrix Human Clariom D Assay | TC0900009905.hg.1 | ENST00000610543.4 | Ensembl |
| Affymetrix Human Clariom D Assay | TC0900009905.hg.1 | NM_001207011.1 | Refseq |
| Affymetrix Human Clariom D Assay | TC0900009905.hg.1 | NM_001207011.1 | Refseq |
| Affymetrix Human Clariom D Assay | TC0900009905.hg.1 | NM_001207011.1 | Refseq |
| Affymetrix Human Clariom D Assay | TC0900009905.hg.1 | NM_001207011.1 | Refseq |
| Affymetrix Human Clariom D Assay | TC0900009905.hg.1 | NM_001207011.1 | Refseq |
| Affymetrix Human Clariom D Assay | TC0900009905.hg.1 | NM_001207011.1 | Refseq |
| Affymetrix Human Clariom D Assay | TC0900009905.hg.1 | NM_001207011.1 | Refseq |
| Affymetrix Human Clariom D Assay | TC0900009905.hg.1 | NM_001207011.1 | Refseq |
| Affymetrix Human Clariom D Assay | TC0900009905.hg.1 | NM_001207011.1 | Refseq |
| Affymetrix Human Clariom D Assay | TC0900009905.hg.1 | NM_001207011.1 | Refseq |
| Affymetrix Human Clariom D Assay | TC0900009905.hg.1 | NM_001842.4 | Refseq |
| Affymetrix Human Clariom D Assay | TC0900009905.hg.1 | NM_001842.4 | Refseq |
| Affymetrix Human Clariom D Assay | TC0900009905.hg.1 | NM_001842.4 | Refseq |
| Affymetrix Human Clariom D Assay | TC0900009905.hg.1 | NM_001842.4 | Refseq |
| Affymetrix Human Clariom D Assay | TC0900009905.hg.1 | NM_001842.4 | Refseq |
| Affymetrix Human Clariom D Assay | TC0900009905.hg.1 | NM_001842.4 | Refseq |
| Affymetrix Human Clariom D Assay | TC0900009905.hg.1 | NM_001842.4 | Refseq |
| Affymetrix Human Clariom D Assay | TC0900009905.hg.1 | NM_001842.4 | Refseq |
| Affymetrix Human Clariom D Assay | TC0900009905.hg.1 | NM_001842.4 | Refseq |
| Affymetrix Human Clariom D Assay | TC0900009905.hg.1 | NM_001842.4 | Refseq |
| Affymetrix Human Clariom D Assay | TC0900009905.hg.1 | NM_147164.2 | Refseq |
| Affymetrix Human Clariom D Assay | TC0900009905.hg.1 | NM_147164.2 | Refseq |
| Affymetrix Human Clariom D Assay | TC0900009905.hg.1 | NM_147164.2 | Refseq |
| Affymetrix Human Clariom D Assay | TC0900009905.hg.1 | NM_147164.2 | Refseq |
| Affymetrix Human Clariom D Assay | TC0900009905.hg.1 | NM_147164.2 | Refseq |
| Affymetrix Human Clariom D Assay | TC0900009905.hg.1 | NM_147164.2 | Refseq |
| Affymetrix Human Clariom D Assay | TC0900009905.hg.1 | NM_147164.2 | Refseq |
| Affymetrix Human Clariom D Assay | TC0900009905.hg.1 | NM_147164.2 | Refseq |
| Affymetrix Human Clariom D Assay | TC0900009905.hg.1 | NM_147164.2 | Refseq |
| Affymetrix Human Clariom D Assay | TC0900009905.hg.1 | NM_147164.2 | Refseq |
| Affymetrix Human Clariom D Assay | TC0900009905.hg.1 | NM_147164.2 | Refseq |
| Affymetrix Human Clariom D Assay | PSR0900167882.hg.1 | ENST00000417345.2 | Ensembl |
| Affymetrix Human Clariom D Assay | PSR0900167882.hg.1 | ENST00000610543.4 | Ensembl |
| Affymetrix Human Clariom D Assay | PSR0900167882.hg.1 | NM_001207011.1 | Refseq |
| Affymetrix Human Clariom D Assay | PSR0900167882.hg.1 | ENST00000417345.2 | Ensembl |
| Affymetrix Human Clariom D Assay | PSR0900167882.hg.1 | ENST00000610543.4 | Ensembl |
| Affymetrix Human Clariom D Assay | PSR0900167882.hg.1 | NM_001207011.1 | Refseq |
| Affymetrix Human Clariom D Assay | PSR0900167882.hg.1 | TC0900009905.hg.1 | Clariom D TC |

CNTFR
Genomic Map