Assay Details
| Gene Symbol: MCTP2 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | NM_001159644.1 | NP_001153116.1 | 1 - 2 | 365 | 94 | |
| XM_005254960.2 | XP_005255017.1 | 1 - 2 | 370 | 94 | ||
| GenBank mRNA | AK002037.1 | - | 1 - 2 | 73 | 94 | |
| AK300325.1 | - | 9 - 10 | 1360 | 94 | ||
| BC025708.1 | - | 1 - 2 | 326 | 94 | ||
| BC111024.1 | - | 1 - 2 | 198 | 94 | ||
More Information
Updated Mapping Information
This assay targets the same location as HTA transcript cluster(s) TC15000930.hg.1
Additional Information:
This assay is in the 5'UTR region of the AK002037.1,BC025708.1,BC111024.1,NM_001159644.1,XM_005254960.2 transcripts.
Set Membership: |
|
Gene Ontology Categories:
MicroArray Probes:
Affymetrix
| Array Name | Probe IDs | Targets | Target Type |
|---|---|---|---|
| Affymetrix Human Transcriptome Array 2.0 | TC1500010792.hg.1 | ENST00000357742.8 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC1500010792.hg.1 | ENST00000357742.8 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC1500010792.hg.1 | ENST00000357742.8 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC1500010792.hg.1 | ENST00000357742.8 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC1500010792.hg.1 | ENST00000357742.8 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC1500010792.hg.1 | ENST00000357742.8 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC1500010792.hg.1 | ENST00000357742.8 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC1500010792.hg.1 | ENST00000357742.8 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC1500010792.hg.1 | ENST00000357742.8 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC1500010792.hg.1 | ENST00000357742.8 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC1500010792.hg.1 | ENST00000357742.8 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC1500010792.hg.1 | ENST00000357742.8 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC1500010792.hg.1 | ENST00000357742.8 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC1500010792.hg.1 | ENST00000357742.8 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC1500010792.hg.1 | ENST00000357742.8 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC1500010792.hg.1 | ENST00000357742.8 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC1500010792.hg.1 | ENST00000357742.8 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC1500010792.hg.1 | ENST00000357742.8 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC1500010792.hg.1 | ENST00000357742.8 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC1500010792.hg.1 | ENST00000357742.8 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC1500010792.hg.1 | ENST00000357742.8 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC1500010792.hg.1 | ENST00000357742.8 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC1500010792.hg.1 | ENST00000357742.8 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC1500010792.hg.1 | ENST00000357742.8 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC1500010792.hg.1 | ENST00000357742.8 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC1500010792.hg.1 | ENST00000357742.8 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC1500010792.hg.1 | ENST00000449432.3 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC1500010792.hg.1 | ENST00000449432.3 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC1500010792.hg.1 | ENST00000449432.3 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC1500010792.hg.1 | ENST00000449432.3 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC1500010792.hg.1 | ENST00000451018.7 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC1500010792.hg.1 | ENST00000451018.7 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC1500010792.hg.1 | ENST00000451018.7 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC1500010792.hg.1 | ENST00000451018.7 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC1500010792.hg.1 | ENST00000451018.7 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC1500010792.hg.1 | ENST00000451018.7 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC1500010792.hg.1 | ENST00000451018.7 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC1500010792.hg.1 | ENST00000451018.7 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC1500010792.hg.1 | ENST00000451018.7 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC1500010792.hg.1 | ENST00000451018.7 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC1500010792.hg.1 | ENST00000451018.7 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC1500010792.hg.1 | ENST00000451018.7 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC1500010792.hg.1 | ENST00000451018.7 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC1500010792.hg.1 | ENST00000451018.7 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC1500010792.hg.1 | ENST00000451018.7 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC1500010792.hg.1 | ENST00000451018.7 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC1500010792.hg.1 | ENST00000451018.7 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC1500010792.hg.1 | ENST00000451018.7 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC1500010792.hg.1 | ENST00000451018.7 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC1500010792.hg.1 | ENST00000451018.7 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC1500010792.hg.1 | ENST00000451018.7 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC1500010792.hg.1 | ENST00000557505.1 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC1500010792.hg.1 | ENST00000557505.1 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC1500010792.hg.1 | ENST00000557505.1 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC1500010792.hg.1 | ENST00000557742.1 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC1500010792.hg.1 | ENST00000557742.1 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC1500010792.hg.1 | ENST00000557742.1 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC1500010792.hg.1 | ENST00000557742.1 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC1500010792.hg.1 | ENST00000557742.1 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC1500010792.hg.1 | ENST00000557742.1 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC1500010792.hg.1 | ENST00000557742.1 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC1500010792.hg.1 | ENST00000557742.1 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC1500010792.hg.1 | ENST00000557742.1 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC1500010792.hg.1 | ENST00000557742.1 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC1500010792.hg.1 | ENST00000557742.1 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC1500010792.hg.1 | ENST00000557742.1 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC1500010792.hg.1 | ENST00000565106.1 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC1500010792.hg.1 | NM_001159643.1 | Refseq |
| Affymetrix Human Transcriptome Array 2.0 | TC1500010792.hg.1 | NM_001159643.1 | Refseq |
| Affymetrix Human Transcriptome Array 2.0 | TC1500010792.hg.1 | NM_001159643.1 | Refseq |
| Affymetrix Human Transcriptome Array 2.0 | TC1500010792.hg.1 | NM_001159643.1 | Refseq |
| Affymetrix Human Transcriptome Array 2.0 | TC1500010792.hg.1 | NM_001159643.1 | Refseq |
| Affymetrix Human Transcriptome Array 2.0 | TC1500010792.hg.1 | NM_001159643.1 | Refseq |
| Affymetrix Human Transcriptome Array 2.0 | TC1500010792.hg.1 | NM_001159643.1 | Refseq |
| Affymetrix Human Transcriptome Array 2.0 | TC1500010792.hg.1 | NM_001159643.1 | Refseq |
| Affymetrix Human Transcriptome Array 2.0 | TC1500010792.hg.1 | NM_001159643.1 | Refseq |
| Affymetrix Human Transcriptome Array 2.0 | TC1500010792.hg.1 | NM_001159643.1 | Refseq |
| Affymetrix Human Transcriptome Array 2.0 | TC1500010792.hg.1 | NM_001159643.1 | Refseq |
| Affymetrix Human Transcriptome Array 2.0 | TC1500010792.hg.1 | NM_001159643.1 | Refseq |
| Affymetrix Human Transcriptome Array 2.0 | TC1500010792.hg.1 | NM_001159643.1 | Refseq |
| Affymetrix Human Transcriptome Array 2.0 | TC1500010792.hg.1 | NM_001159643.1 | Refseq |
| Affymetrix Human Transcriptome Array 2.0 | TC1500010792.hg.1 | NM_001159643.1 | Refseq |
| Affymetrix Human Transcriptome Array 2.0 | TC1500010792.hg.1 | NM_001159643.1 | Refseq |
| Affymetrix Human Transcriptome Array 2.0 | TC1500010792.hg.1 | NM_001159643.1 | Refseq |
| Affymetrix Human Transcriptome Array 2.0 | TC1500010792.hg.1 | NM_001159643.1 | Refseq |
| Affymetrix Human Transcriptome Array 2.0 | TC1500010792.hg.1 | NM_001159643.1 | Refseq |
| Affymetrix Human Transcriptome Array 2.0 | TC1500010792.hg.1 | NM_001159643.1 | Refseq |
| Affymetrix Human Transcriptome Array 2.0 | TC1500010792.hg.1 | NM_001159643.1 | Refseq |
| Affymetrix Human Transcriptome Array 2.0 | TC1500010792.hg.1 | NM_001159643.1 | Refseq |
| Affymetrix Human Transcriptome Array 2.0 | TC1500010792.hg.1 | NM_001159643.1 | Refseq |
| Affymetrix Human Transcriptome Array 2.0 | TC1500010792.hg.1 | NM_001159643.1 | Refseq |
| Affymetrix Human Transcriptome Array 2.0 | TC1500010792.hg.1 | NM_001159643.1 | Refseq |
| Affymetrix Human Transcriptome Array 2.0 | TC1500010792.hg.1 | NM_001159644.1 | Refseq |
| Affymetrix Human Transcriptome Array 2.0 | TC1500010792.hg.1 | NM_001159644.1 | Refseq |
| Affymetrix Human Transcriptome Array 2.0 | TC1500010792.hg.1 | NM_001159644.1 | Refseq |
| Affymetrix Human Transcriptome Array 2.0 | TC1500010792.hg.1 | NM_001159644.1 | Refseq |
| Affymetrix Human Transcriptome Array 2.0 | TC1500010792.hg.1 | NM_001159644.1 | Refseq |
| Affymetrix Human Transcriptome Array 2.0 | TC1500010792.hg.1 | NM_001159644.1 | Refseq |
| Affymetrix Human Transcriptome Array 2.0 | TC1500010792.hg.1 | NM_001159644.1 | Refseq |
| Affymetrix Human Transcriptome Array 2.0 | TC1500010792.hg.1 | NM_001159644.1 | Refseq |
| Affymetrix Human Transcriptome Array 2.0 | TC1500010792.hg.1 | NM_001159644.1 | Refseq |
| Affymetrix Human Transcriptome Array 2.0 | TC1500010792.hg.1 | NM_001159644.1 | Refseq |
| Affymetrix Human Transcriptome Array 2.0 | TC1500010792.hg.1 | NM_001159644.1 | Refseq |
| Affymetrix Human Transcriptome Array 2.0 | TC1500010792.hg.1 | NM_001159644.1 | Refseq |
| Affymetrix Human Transcriptome Array 2.0 | TC1500010792.hg.1 | NM_001159644.1 | Refseq |
| Affymetrix Human Transcriptome Array 2.0 | TC1500010792.hg.1 | NM_018349.3 | Refseq |
| Affymetrix Human Transcriptome Array 2.0 | TC1500010792.hg.1 | NM_018349.3 | Refseq |
| Affymetrix Human Transcriptome Array 2.0 | TC1500010792.hg.1 | NM_018349.3 | Refseq |
| Affymetrix Human Transcriptome Array 2.0 | TC1500010792.hg.1 | NM_018349.3 | Refseq |
| Affymetrix Human Transcriptome Array 2.0 | TC1500010792.hg.1 | NM_018349.3 | Refseq |
| Affymetrix Human Transcriptome Array 2.0 | TC1500010792.hg.1 | NM_018349.3 | Refseq |
| Affymetrix Human Transcriptome Array 2.0 | TC1500010792.hg.1 | NM_018349.3 | Refseq |
| Affymetrix Human Transcriptome Array 2.0 | TC1500010792.hg.1 | NM_018349.3 | Refseq |
| Affymetrix Human Transcriptome Array 2.0 | TC1500010792.hg.1 | NM_018349.3 | Refseq |
| Affymetrix Human Transcriptome Array 2.0 | TC1500010792.hg.1 | NM_018349.3 | Refseq |
| Affymetrix Human Transcriptome Array 2.0 | TC1500010792.hg.1 | NM_018349.3 | Refseq |
| Affymetrix Human Transcriptome Array 2.0 | TC1500010792.hg.1 | NM_018349.3 | Refseq |
| Affymetrix Human Transcriptome Array 2.0 | TC1500010792.hg.1 | NM_018349.3 | Refseq |
| Affymetrix Human Transcriptome Array 2.0 | TC1500010792.hg.1 | NM_018349.3 | Refseq |
| Affymetrix Human Transcriptome Array 2.0 | TC1500010792.hg.1 | NM_018349.3 | Refseq |
| Affymetrix Human Transcriptome Array 2.0 | TC1500010792.hg.1 | NM_018349.3 | Refseq |
| Affymetrix Human Transcriptome Array 2.0 | TC1500010792.hg.1 | NM_018349.3 | Refseq |
| Affymetrix Human Transcriptome Array 2.0 | TC1500010792.hg.1 | NM_018349.3 | Refseq |
| Affymetrix Human Transcriptome Array 2.0 | TC1500010792.hg.1 | NM_018349.3 | Refseq |
| Affymetrix Human Transcriptome Array 2.0 | TC1500010792.hg.1 | NM_018349.3 | Refseq |
| Affymetrix Human Transcriptome Array 2.0 | TC1500010792.hg.1 | NM_018349.3 | Refseq |
| Affymetrix Human Transcriptome Array 2.0 | TC1500010792.hg.1 | NM_018349.3 | Refseq |
| Affymetrix Human Transcriptome Array 2.0 | TC1500010792.hg.1 | NM_018349.3 | Refseq |
| Affymetrix Human Transcriptome Array 2.0 | TC1500010792.hg.1 | NM_018349.3 | Refseq |
| Affymetrix Human Transcriptome Array 2.0 | TC1500010792.hg.1 | NM_018349.3 | Refseq |
| Affymetrix Human Transcriptome Array 2.0 | TC1500010792.hg.1 | NM_018349.3 | Refseq |
| Affymetrix Human Transcriptome Array 2.0 | TC1500010792.hg.1 | NM_018349.3 | Refseq |

MCTP2
Genomic Map