Assay Details
| Gene Symbol: EIF4H | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | NM_022170.1 | NP_071496.1 | 4 - 5 | 412 | 110 | |
| XM_011516556.1 | XP_011514858.1 | 4 - 5 | 445 | 110 | ||
More Information
Updated Mapping Information
This assay targets the same location as HTA transcript cluster(s) TC07000451.hg.1
Set Membership: |
|
Matching Ambion siRNAs:
Panther Classification:
Gene Ontology Categories:
Function(s)
Process(es)
formation of translation preinitiation complex
cytoplasmic translation
translational initiation
regulation of translational initiation
viral process
sexual reproduction
developmental growth
eukaryotic translation initiation factor 4F complex assembly
cell-cell adhesion
nucleotide binding
RNA binding
translation initiation factor activity
helicase activity
protein binding
translation factor activity, RNA binding
RNA strand annealing activity
RNA strand-exchange activity
ribosomal small subunit binding
poly(A) RNA binding
cadherin binding involved in cell-cell adhesion
formation of translation preinitiation complex
cytoplasmic translation
translational initiation
regulation of translational initiation
viral process
sexual reproduction
developmental growth
eukaryotic translation initiation factor 4F complex assembly
cell-cell adhesion
nucleotide binding
RNA binding
translation initiation factor activity
helicase activity
protein binding
translation factor activity, RNA binding
RNA strand annealing activity
RNA strand-exchange activity
ribosomal small subunit binding
poly(A) RNA binding
cadherin binding involved in cell-cell adhesion
MicroArray Probes:
Affymetrix
| Array Name | Probe IDs | Targets | Target Type |
|---|---|---|---|
| Affymetrix Human Transcriptome Array 2.0 | PSR07006110.hg.1 | TC07000451.hg.1 | HTA TC |
| Affymetrix Human Transcriptome Array 2.0 | PSR07006110.hg.1 | ENST00000265753.8 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | PSR07006110.hg.1 | ENST00000495187.1 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | PSR07006110.hg.1 | NM_022170.1 | Refseq |
| Affymetrix Human Transcriptome Array 2.0 | TC0700007998.hg.1 | ENST00000265753.12 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0700007998.hg.1 | ENST00000265753.12 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0700007998.hg.1 | ENST00000265753.12 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0700007998.hg.1 | ENST00000265753.12 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0700007998.hg.1 | ENST00000265753.12 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0700007998.hg.1 | ENST00000265753.12 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0700007998.hg.1 | ENST00000265753.12 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0700007998.hg.1 | ENST00000265753.12 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0700007998.hg.1 | ENST00000265753.12 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0700007998.hg.1 | ENST00000265753.12 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0700007998.hg.1 | ENST00000265753.12 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0700007998.hg.1 | ENST00000353999.6 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0700007998.hg.1 | ENST00000353999.6 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0700007998.hg.1 | ENST00000353999.6 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0700007998.hg.1 | ENST00000353999.6 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0700007998.hg.1 | ENST00000353999.6 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0700007998.hg.1 | ENST00000353999.6 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0700007998.hg.1 | ENST00000353999.6 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0700007998.hg.1 | ENST00000353999.6 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0700007998.hg.1 | ENST00000479338.5 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0700007998.hg.1 | ENST00000479338.5 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0700007998.hg.1 | ENST00000479338.5 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0700007998.hg.1 | ENST00000479338.5 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0700007998.hg.1 | ENST00000479338.5 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0700007998.hg.1 | ENST00000479338.5 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0700007998.hg.1 | ENST00000489484.5 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0700007998.hg.1 | ENST00000489484.5 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0700007998.hg.1 | ENST00000489484.5 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0700007998.hg.1 | ENST00000489484.5 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0700007998.hg.1 | ENST00000489484.5 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0700007998.hg.1 | ENST00000489484.5 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0700007998.hg.1 | ENST00000489484.5 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0700007998.hg.1 | ENST00000489484.5 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0700007998.hg.1 | ENST00000495187.1 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0700007998.hg.1 | ENST00000495187.1 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0700007998.hg.1 | ENST00000495187.1 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0700007998.hg.1 | ENST00000495187.1 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0700007998.hg.1 | ENST00000495187.1 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0700007998.hg.1 | ENST00000495187.1 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0700007998.hg.1 | NM_022170.1 | Refseq |
| Affymetrix Human Transcriptome Array 2.0 | TC0700007998.hg.1 | NM_022170.1 | Refseq |
| Affymetrix Human Transcriptome Array 2.0 | TC0700007998.hg.1 | NM_022170.1 | Refseq |
| Affymetrix Human Transcriptome Array 2.0 | TC0700007998.hg.1 | NM_022170.1 | Refseq |
| Affymetrix Human Transcriptome Array 2.0 | TC0700007998.hg.1 | NM_022170.1 | Refseq |
| Affymetrix Human Transcriptome Array 2.0 | TC0700007998.hg.1 | NM_022170.1 | Refseq |
| Affymetrix Human Transcriptome Array 2.0 | TC0700007998.hg.1 | NM_022170.1 | Refseq |
| Affymetrix Human Transcriptome Array 2.0 | TC0700007998.hg.1 | NM_022170.1 | Refseq |
| Affymetrix Human Transcriptome Array 2.0 | TC0700007998.hg.1 | NM_022170.1 | Refseq |
| Affymetrix Human Transcriptome Array 2.0 | TC0700007998.hg.1 | NM_031992.1 | Refseq |
| Affymetrix Human Transcriptome Array 2.0 | TC0700007998.hg.1 | NM_031992.1 | Refseq |
| Affymetrix Human Transcriptome Array 2.0 | TC0700007998.hg.1 | NM_031992.1 | Refseq |
| Affymetrix Human Transcriptome Array 2.0 | TC0700007998.hg.1 | NM_031992.1 | Refseq |
| Affymetrix Human Transcriptome Array 2.0 | TC0700007998.hg.1 | NM_031992.1 | Refseq |
| Affymetrix Human Transcriptome Array 2.0 | TC0700007998.hg.1 | NM_031992.1 | Refseq |
| Affymetrix Human Transcriptome Array 2.0 | TC0700007998.hg.1 | NM_031992.1 | Refseq |
| Affymetrix Human Transcriptome Array 2.0 | TC0700007998.hg.1 | NM_031992.1 | Refseq |
| Affymetrix Human Clariom D Assay | PSR0700155242.hg.1 | ENST00000265753.12 | Ensembl |
| Affymetrix Human Clariom D Assay | PSR0700155242.hg.1 | ENST00000489484.5 | Ensembl |
| Affymetrix Human Clariom D Assay | PSR0700155242.hg.1 | ENST00000495187.1 | Ensembl |
| Affymetrix Human Clariom D Assay | PSR0700155242.hg.1 | NM_022170.1 | Refseq |
| Affymetrix Human Clariom D Assay | PSR0700155242.hg.1 | ENST00000265753.12 | Ensembl |
| Affymetrix Human Clariom D Assay | PSR0700155242.hg.1 | ENST00000489484.5 | Ensembl |
| Affymetrix Human Clariom D Assay | PSR0700155242.hg.1 | ENST00000495187.1 | Ensembl |
| Affymetrix Human Clariom D Assay | PSR0700155242.hg.1 | NM_022170.1 | Refseq |
| Affymetrix Human Clariom D Assay | PSR0700155242.hg.1 | TC0700007998.hg.1 | Clariom D TC |
| Affymetrix Human Clariom D Assay | TC0700007998.hg.1 | ENST00000265753.12 | Ensembl |
| Affymetrix Human Clariom D Assay | TC0700007998.hg.1 | ENST00000265753.12 | Ensembl |
| Affymetrix Human Clariom D Assay | TC0700007998.hg.1 | ENST00000265753.12 | Ensembl |
| Affymetrix Human Clariom D Assay | TC0700007998.hg.1 | ENST00000265753.12 | Ensembl |
| Affymetrix Human Clariom D Assay | TC0700007998.hg.1 | ENST00000265753.12 | Ensembl |
| Affymetrix Human Clariom D Assay | TC0700007998.hg.1 | ENST00000265753.12 | Ensembl |
| Affymetrix Human Clariom D Assay | TC0700007998.hg.1 | ENST00000265753.12 | Ensembl |
| Affymetrix Human Clariom D Assay | TC0700007998.hg.1 | ENST00000265753.12 | Ensembl |
| Affymetrix Human Clariom D Assay | TC0700007998.hg.1 | ENST00000265753.12 | Ensembl |
| Affymetrix Human Clariom D Assay | TC0700007998.hg.1 | ENST00000265753.12 | Ensembl |
| Affymetrix Human Clariom D Assay | TC0700007998.hg.1 | ENST00000265753.12 | Ensembl |
| Affymetrix Human Clariom D Assay | TC0700007998.hg.1 | ENST00000353999.6 | Ensembl |
| Affymetrix Human Clariom D Assay | TC0700007998.hg.1 | ENST00000353999.6 | Ensembl |
| Affymetrix Human Clariom D Assay | TC0700007998.hg.1 | ENST00000353999.6 | Ensembl |
| Affymetrix Human Clariom D Assay | TC0700007998.hg.1 | ENST00000353999.6 | Ensembl |
| Affymetrix Human Clariom D Assay | TC0700007998.hg.1 | ENST00000353999.6 | Ensembl |
| Affymetrix Human Clariom D Assay | TC0700007998.hg.1 | ENST00000353999.6 | Ensembl |
| Affymetrix Human Clariom D Assay | TC0700007998.hg.1 | ENST00000353999.6 | Ensembl |
| Affymetrix Human Clariom D Assay | TC0700007998.hg.1 | ENST00000353999.6 | Ensembl |
| Affymetrix Human Clariom D Assay | TC0700007998.hg.1 | ENST00000479338.5 | Ensembl |
| Affymetrix Human Clariom D Assay | TC0700007998.hg.1 | ENST00000479338.5 | Ensembl |
| Affymetrix Human Clariom D Assay | TC0700007998.hg.1 | ENST00000479338.5 | Ensembl |
| Affymetrix Human Clariom D Assay | TC0700007998.hg.1 | ENST00000479338.5 | Ensembl |
| Affymetrix Human Clariom D Assay | TC0700007998.hg.1 | ENST00000479338.5 | Ensembl |
| Affymetrix Human Clariom D Assay | TC0700007998.hg.1 | ENST00000479338.5 | Ensembl |
| Affymetrix Human Clariom D Assay | TC0700007998.hg.1 | ENST00000489484.5 | Ensembl |
| Affymetrix Human Clariom D Assay | TC0700007998.hg.1 | ENST00000489484.5 | Ensembl |
| Affymetrix Human Clariom D Assay | TC0700007998.hg.1 | ENST00000489484.5 | Ensembl |
| Affymetrix Human Clariom D Assay | TC0700007998.hg.1 | ENST00000489484.5 | Ensembl |
| Affymetrix Human Clariom D Assay | TC0700007998.hg.1 | ENST00000489484.5 | Ensembl |
| Affymetrix Human Clariom D Assay | TC0700007998.hg.1 | ENST00000489484.5 | Ensembl |
| Affymetrix Human Clariom D Assay | TC0700007998.hg.1 | ENST00000489484.5 | Ensembl |
| Affymetrix Human Clariom D Assay | TC0700007998.hg.1 | ENST00000489484.5 | Ensembl |
| Affymetrix Human Clariom D Assay | TC0700007998.hg.1 | ENST00000495187.1 | Ensembl |
| Affymetrix Human Clariom D Assay | TC0700007998.hg.1 | ENST00000495187.1 | Ensembl |
| Affymetrix Human Clariom D Assay | TC0700007998.hg.1 | ENST00000495187.1 | Ensembl |
| Affymetrix Human Clariom D Assay | TC0700007998.hg.1 | ENST00000495187.1 | Ensembl |
| Affymetrix Human Clariom D Assay | TC0700007998.hg.1 | ENST00000495187.1 | Ensembl |
| Affymetrix Human Clariom D Assay | TC0700007998.hg.1 | ENST00000495187.1 | Ensembl |
| Affymetrix Human Clariom D Assay | TC0700007998.hg.1 | NM_022170.1 | Refseq |
| Affymetrix Human Clariom D Assay | TC0700007998.hg.1 | NM_022170.1 | Refseq |
| Affymetrix Human Clariom D Assay | TC0700007998.hg.1 | NM_022170.1 | Refseq |
| Affymetrix Human Clariom D Assay | TC0700007998.hg.1 | NM_022170.1 | Refseq |
| Affymetrix Human Clariom D Assay | TC0700007998.hg.1 | NM_022170.1 | Refseq |
| Affymetrix Human Clariom D Assay | TC0700007998.hg.1 | NM_022170.1 | Refseq |
| Affymetrix Human Clariom D Assay | TC0700007998.hg.1 | NM_022170.1 | Refseq |
| Affymetrix Human Clariom D Assay | TC0700007998.hg.1 | NM_022170.1 | Refseq |
| Affymetrix Human Clariom D Assay | TC0700007998.hg.1 | NM_022170.1 | Refseq |
| Affymetrix Human Clariom D Assay | TC0700007998.hg.1 | NM_031992.1 | Refseq |
| Affymetrix Human Clariom D Assay | TC0700007998.hg.1 | NM_031992.1 | Refseq |
| Affymetrix Human Clariom D Assay | TC0700007998.hg.1 | NM_031992.1 | Refseq |
| Affymetrix Human Clariom D Assay | TC0700007998.hg.1 | NM_031992.1 | Refseq |
| Affymetrix Human Clariom D Assay | TC0700007998.hg.1 | NM_031992.1 | Refseq |
| Affymetrix Human Clariom D Assay | TC0700007998.hg.1 | NM_031992.1 | Refseq |
| Affymetrix Human Clariom D Assay | TC0700007998.hg.1 | NM_031992.1 | Refseq |
| Affymetrix Human Clariom D Assay | TC0700007998.hg.1 | NM_031992.1 | Refseq |

EIF4H
Genomic Map