Assay Details
| Gene Symbol: MRPL34 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | XM_011528189.2 | XP_011526491.1 | 1 - 2 | 135 | 67 | |
More Information
Additional Information:
This assay is in the 5'UTR region of the XM_011528189.2 transcripts.
Based on assay re-evaluation, this assay may detect genomic DNA. Please contact Tech Support if you need more information.
Based on assay re-evaluation, this assay may detect genomic DNA. Please contact Tech Support if you need more information.
Set Membership: |
|
Panther Classification:
Gene Ontology Categories:
MicroArray Probes:
Affymetrix
| Array Name | Probe IDs | Targets | Target Type |
|---|---|---|---|
| Affymetrix Human Transcriptome Array 2.0 | TC1900007319.hg.1 | ENST00000252602.1 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC1900007319.hg.1 | ENST00000252602.1 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC1900007319.hg.1 | ENST00000252602.1 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC1900007319.hg.1 | ENST00000252602.1 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC1900007319.hg.1 | ENST00000252602.1 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC1900007319.hg.1 | ENST00000252602.1 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC1900007319.hg.1 | ENST00000252602.1 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC1900007319.hg.1 | ENST00000252602.1 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC1900007319.hg.1 | ENST00000252602.1 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC1900007319.hg.1 | ENST00000594999.1 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC1900007319.hg.1 | ENST00000594999.1 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC1900007319.hg.1 | ENST00000594999.1 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC1900007319.hg.1 | ENST00000594999.1 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC1900007319.hg.1 | ENST00000594999.1 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC1900007319.hg.1 | ENST00000594999.1 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC1900007319.hg.1 | ENST00000595444.1 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC1900007319.hg.1 | ENST00000595444.1 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC1900007319.hg.1 | ENST00000595444.1 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC1900007319.hg.1 | ENST00000595444.1 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC1900007319.hg.1 | ENST00000595444.1 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC1900007319.hg.1 | ENST00000595444.1 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC1900007319.hg.1 | ENST00000595444.1 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC1900007319.hg.1 | ENST00000597996.1 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC1900007319.hg.1 | ENST00000597996.1 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC1900007319.hg.1 | ENST00000597996.1 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC1900007319.hg.1 | ENST00000597996.1 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC1900007319.hg.1 | ENST00000600434.5 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC1900007319.hg.1 | ENST00000600434.5 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC1900007319.hg.1 | ENST00000600434.5 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC1900007319.hg.1 | ENST00000600434.5 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC1900007319.hg.1 | ENST00000600434.5 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC1900007319.hg.1 | ENST00000600434.5 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC1900007319.hg.1 | ENST00000600434.5 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC1900007319.hg.1 | ENST00000602206.1 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC1900007319.hg.1 | ENST00000602206.1 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC1900007319.hg.1 | ENST00000602206.1 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC1900007319.hg.1 | ENST00000602206.1 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC1900007319.hg.1 | ENST00000602206.1 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC1900007319.hg.1 | NM_023937.3 | Refseq |
| Affymetrix Human Transcriptome Array 2.0 | TC1900007319.hg.1 | NM_023937.3 | Refseq |
| Affymetrix Human Transcriptome Array 2.0 | TC1900007319.hg.1 | NM_023937.3 | Refseq |
| Affymetrix Human Transcriptome Array 2.0 | TC1900007319.hg.1 | NM_023937.3 | Refseq |
| Affymetrix Human Transcriptome Array 2.0 | TC1900007319.hg.1 | NM_023937.3 | Refseq |
| Affymetrix Human Transcriptome Array 2.0 | TC1900007319.hg.1 | NM_023937.3 | Refseq |
| Affymetrix Human Transcriptome Array 2.0 | TC1900007319.hg.1 | NM_023937.3 | Refseq |
| Affymetrix Human Transcriptome Array 2.0 | TC1900007319.hg.1 | NM_023937.3 | Refseq |
| Affymetrix Human Transcriptome Array 2.0 | TC1900007319.hg.1 | NM_023937.3 | Refseq |
| Affymetrix Human Clariom D Assay | TC1900007319.hg.1 | ENST00000252602.1 | Ensembl |
| Affymetrix Human Clariom D Assay | TC1900007319.hg.1 | ENST00000252602.1 | Ensembl |
| Affymetrix Human Clariom D Assay | TC1900007319.hg.1 | ENST00000252602.1 | Ensembl |
| Affymetrix Human Clariom D Assay | TC1900007319.hg.1 | ENST00000252602.1 | Ensembl |
| Affymetrix Human Clariom D Assay | TC1900007319.hg.1 | ENST00000252602.1 | Ensembl |
| Affymetrix Human Clariom D Assay | TC1900007319.hg.1 | ENST00000252602.1 | Ensembl |
| Affymetrix Human Clariom D Assay | TC1900007319.hg.1 | ENST00000252602.1 | Ensembl |
| Affymetrix Human Clariom D Assay | TC1900007319.hg.1 | ENST00000252602.1 | Ensembl |
| Affymetrix Human Clariom D Assay | TC1900007319.hg.1 | ENST00000252602.1 | Ensembl |
| Affymetrix Human Clariom D Assay | TC1900007319.hg.1 | ENST00000594999.1 | Ensembl |
| Affymetrix Human Clariom D Assay | TC1900007319.hg.1 | ENST00000594999.1 | Ensembl |
| Affymetrix Human Clariom D Assay | TC1900007319.hg.1 | ENST00000594999.1 | Ensembl |
| Affymetrix Human Clariom D Assay | TC1900007319.hg.1 | ENST00000594999.1 | Ensembl |
| Affymetrix Human Clariom D Assay | TC1900007319.hg.1 | ENST00000594999.1 | Ensembl |
| Affymetrix Human Clariom D Assay | TC1900007319.hg.1 | ENST00000594999.1 | Ensembl |
| Affymetrix Human Clariom D Assay | TC1900007319.hg.1 | ENST00000595444.1 | Ensembl |
| Affymetrix Human Clariom D Assay | TC1900007319.hg.1 | ENST00000595444.1 | Ensembl |
| Affymetrix Human Clariom D Assay | TC1900007319.hg.1 | ENST00000595444.1 | Ensembl |
| Affymetrix Human Clariom D Assay | TC1900007319.hg.1 | ENST00000595444.1 | Ensembl |
| Affymetrix Human Clariom D Assay | TC1900007319.hg.1 | ENST00000595444.1 | Ensembl |
| Affymetrix Human Clariom D Assay | TC1900007319.hg.1 | ENST00000595444.1 | Ensembl |
| Affymetrix Human Clariom D Assay | TC1900007319.hg.1 | ENST00000595444.1 | Ensembl |
| Affymetrix Human Clariom D Assay | TC1900007319.hg.1 | ENST00000597996.1 | Ensembl |
| Affymetrix Human Clariom D Assay | TC1900007319.hg.1 | ENST00000597996.1 | Ensembl |
| Affymetrix Human Clariom D Assay | TC1900007319.hg.1 | ENST00000597996.1 | Ensembl |
| Affymetrix Human Clariom D Assay | TC1900007319.hg.1 | ENST00000597996.1 | Ensembl |
| Affymetrix Human Clariom D Assay | TC1900007319.hg.1 | ENST00000600434.5 | Ensembl |
| Affymetrix Human Clariom D Assay | TC1900007319.hg.1 | ENST00000600434.5 | Ensembl |
| Affymetrix Human Clariom D Assay | TC1900007319.hg.1 | ENST00000600434.5 | Ensembl |
| Affymetrix Human Clariom D Assay | TC1900007319.hg.1 | ENST00000600434.5 | Ensembl |
| Affymetrix Human Clariom D Assay | TC1900007319.hg.1 | ENST00000600434.5 | Ensembl |
| Affymetrix Human Clariom D Assay | TC1900007319.hg.1 | ENST00000600434.5 | Ensembl |
| Affymetrix Human Clariom D Assay | TC1900007319.hg.1 | ENST00000600434.5 | Ensembl |
| Affymetrix Human Clariom D Assay | TC1900007319.hg.1 | ENST00000602206.1 | Ensembl |
| Affymetrix Human Clariom D Assay | TC1900007319.hg.1 | ENST00000602206.1 | Ensembl |
| Affymetrix Human Clariom D Assay | TC1900007319.hg.1 | ENST00000602206.1 | Ensembl |
| Affymetrix Human Clariom D Assay | TC1900007319.hg.1 | ENST00000602206.1 | Ensembl |
| Affymetrix Human Clariom D Assay | TC1900007319.hg.1 | ENST00000602206.1 | Ensembl |
| Affymetrix Human Clariom D Assay | TC1900007319.hg.1 | NM_023937.3 | Refseq |
| Affymetrix Human Clariom D Assay | TC1900007319.hg.1 | NM_023937.3 | Refseq |
| Affymetrix Human Clariom D Assay | TC1900007319.hg.1 | NM_023937.3 | Refseq |
| Affymetrix Human Clariom D Assay | TC1900007319.hg.1 | NM_023937.3 | Refseq |
| Affymetrix Human Clariom D Assay | TC1900007319.hg.1 | NM_023937.3 | Refseq |
| Affymetrix Human Clariom D Assay | TC1900007319.hg.1 | NM_023937.3 | Refseq |
| Affymetrix Human Clariom D Assay | TC1900007319.hg.1 | NM_023937.3 | Refseq |
| Affymetrix Human Clariom D Assay | TC1900007319.hg.1 | NM_023937.3 | Refseq |
| Affymetrix Human Clariom D Assay | TC1900007319.hg.1 | NM_023937.3 | Refseq |

MRPL34
Genomic Map