Assay Details
| Gene Symbol: RBM20 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | NM_001134363.2 | NP_001127835.2 | 6 - 7 | 1726 | 92 | |
| XM_011539697.2 | XP_011537999.1 | 6 - 7 | 1564 | 92 | ||
| XM_017016103.1 | XP_016871592.1 | 6 - 7 | 2984 | 92 | ||
| XM_017016104.1 | XP_016871593.1 | 6 - 7 | 5200 | 92 | ||
| GenBank mRNA | EU822950.1 | - | 6 - 7 | 1726 | 92 | |
More Information
Updated Mapping Information
This assay targets the same location as HTA transcript cluster(s) TC10000804.hg.1
Set Membership: |
|
Gene Ontology Categories:
MicroArray Probes:
Affymetrix
| Array Name | Probe IDs | Targets | Target Type |
|---|---|---|---|
| Affymetrix Human Transcriptome Array 2.0 | PSR10009635.hg.1 | ENST00000369519.3 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | PSR10009635.hg.1 | ENST00000539821.1 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | PSR10009635.hg.1 | NM_001134363.2 | Refseq |
| Affymetrix Human Transcriptome Array 2.0 | PSR10009635.hg.1 | TC10000804.hg.1 | HTA TC |
| Affymetrix Human Transcriptome Array 2.0 | TC1000008898.hg.1 | ENST00000369519.3 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC1000008898.hg.1 | ENST00000369519.3 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC1000008898.hg.1 | ENST00000369519.3 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC1000008898.hg.1 | ENST00000369519.3 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC1000008898.hg.1 | ENST00000369519.3 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC1000008898.hg.1 | ENST00000369519.3 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC1000008898.hg.1 | ENST00000369519.3 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC1000008898.hg.1 | ENST00000369519.3 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC1000008898.hg.1 | ENST00000369519.3 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC1000008898.hg.1 | ENST00000369519.3 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC1000008898.hg.1 | ENST00000369519.3 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC1000008898.hg.1 | ENST00000369519.3 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC1000008898.hg.1 | ENST00000369519.3 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC1000008898.hg.1 | ENST00000369519.3 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC1000008898.hg.1 | ENST00000369519.3 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC1000008898.hg.1 | ENST00000369519.3 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC1000008898.hg.1 | ENST00000369519.3 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC1000008898.hg.1 | ENST00000369519.3 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC1000008898.hg.1 | ENST00000465774.2 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC1000008898.hg.1 | ENST00000465774.2 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC1000008898.hg.1 | ENST00000465774.2 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC1000008898.hg.1 | ENST00000471172.1 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC1000008898.hg.1 | ENST00000471172.1 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC1000008898.hg.1 | ENST00000471172.1 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC1000008898.hg.1 | ENST00000480343.2 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC1000008898.hg.1 | ENST00000480343.2 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC1000008898.hg.1 | ENST00000480343.2 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC1000008898.hg.1 | ENST00000480343.2 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC1000008898.hg.1 | NM_001134363.2 | Refseq |
| Affymetrix Human Transcriptome Array 2.0 | TC1000008898.hg.1 | NM_001134363.2 | Refseq |
| Affymetrix Human Transcriptome Array 2.0 | TC1000008898.hg.1 | NM_001134363.2 | Refseq |
| Affymetrix Human Transcriptome Array 2.0 | TC1000008898.hg.1 | NM_001134363.2 | Refseq |
| Affymetrix Human Transcriptome Array 2.0 | TC1000008898.hg.1 | NM_001134363.2 | Refseq |
| Affymetrix Human Transcriptome Array 2.0 | TC1000008898.hg.1 | NM_001134363.2 | Refseq |
| Affymetrix Human Transcriptome Array 2.0 | TC1000008898.hg.1 | NM_001134363.2 | Refseq |
| Affymetrix Human Transcriptome Array 2.0 | TC1000008898.hg.1 | NM_001134363.2 | Refseq |
| Affymetrix Human Transcriptome Array 2.0 | TC1000008898.hg.1 | NM_001134363.2 | Refseq |
| Affymetrix Human Transcriptome Array 2.0 | TC1000008898.hg.1 | NM_001134363.2 | Refseq |
| Affymetrix Human Transcriptome Array 2.0 | TC1000008898.hg.1 | NM_001134363.2 | Refseq |
| Affymetrix Human Transcriptome Array 2.0 | TC1000008898.hg.1 | NM_001134363.2 | Refseq |
| Affymetrix Human Transcriptome Array 2.0 | TC1000008898.hg.1 | NM_001134363.2 | Refseq |
| Affymetrix Human Transcriptome Array 2.0 | TC1000008898.hg.1 | NM_001134363.2 | Refseq |
| Affymetrix Human Transcriptome Array 2.0 | TC1000008898.hg.1 | NM_001134363.2 | Refseq |
| Affymetrix Human Transcriptome Array 2.0 | TC1000008898.hg.1 | NM_001134363.2 | Refseq |
| Affymetrix Human Transcriptome Array 2.0 | TC1000008898.hg.1 | NM_001134363.2 | Refseq |
| Affymetrix Human Transcriptome Array 2.0 | TC1000008898.hg.1 | NM_001134363.2 | Refseq |
| Affymetrix Human Transcriptome Array 2.0 | PSR10009636.hg.1 | ENST00000369519.3 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | PSR10009636.hg.1 | ENST00000539821.1 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | PSR10009636.hg.1 | NM_001134363.2 | Refseq |
| Affymetrix Human Transcriptome Array 2.0 | PSR10009636.hg.1 | TC10000804.hg.1 | HTA TC |
| Affymetrix Human Clariom D Assay | PSR1000160840.hg.1 | ENST00000369519.3 | Ensembl |
| Affymetrix Human Clariom D Assay | PSR1000160840.hg.1 | NM_001134363.2 | Refseq |
| Affymetrix Human Clariom D Assay | PSR1000160840.hg.1 | ENST00000369519.3 | Ensembl |
| Affymetrix Human Clariom D Assay | PSR1000160840.hg.1 | NM_001134363.2 | Refseq |
| Affymetrix Human Clariom D Assay | PSR1000160840.hg.1 | TC1000008898.hg.1 | Clariom D TC |
| Affymetrix Human Clariom D Assay | PSR1000160841.hg.1 | ENST00000369519.3 | Ensembl |
| Affymetrix Human Clariom D Assay | PSR1000160841.hg.1 | NM_001134363.2 | Refseq |
| Affymetrix Human Clariom D Assay | PSR1000160841.hg.1 | ENST00000369519.3 | Ensembl |
| Affymetrix Human Clariom D Assay | PSR1000160841.hg.1 | NM_001134363.2 | Refseq |
| Affymetrix Human Clariom D Assay | PSR1000160841.hg.1 | TC1000008898.hg.1 | Clariom D TC |
| Affymetrix Human Clariom D Assay | TC1000008898.hg.1 | ENST00000369519.3 | Ensembl |
| Affymetrix Human Clariom D Assay | TC1000008898.hg.1 | ENST00000369519.3 | Ensembl |
| Affymetrix Human Clariom D Assay | TC1000008898.hg.1 | ENST00000369519.3 | Ensembl |
| Affymetrix Human Clariom D Assay | TC1000008898.hg.1 | ENST00000369519.3 | Ensembl |
| Affymetrix Human Clariom D Assay | TC1000008898.hg.1 | ENST00000369519.3 | Ensembl |
| Affymetrix Human Clariom D Assay | TC1000008898.hg.1 | ENST00000369519.3 | Ensembl |
| Affymetrix Human Clariom D Assay | TC1000008898.hg.1 | ENST00000369519.3 | Ensembl |
| Affymetrix Human Clariom D Assay | TC1000008898.hg.1 | ENST00000369519.3 | Ensembl |
| Affymetrix Human Clariom D Assay | TC1000008898.hg.1 | ENST00000369519.3 | Ensembl |
| Affymetrix Human Clariom D Assay | TC1000008898.hg.1 | ENST00000369519.3 | Ensembl |
| Affymetrix Human Clariom D Assay | TC1000008898.hg.1 | ENST00000369519.3 | Ensembl |
| Affymetrix Human Clariom D Assay | TC1000008898.hg.1 | ENST00000369519.3 | Ensembl |
| Affymetrix Human Clariom D Assay | TC1000008898.hg.1 | ENST00000369519.3 | Ensembl |
| Affymetrix Human Clariom D Assay | TC1000008898.hg.1 | ENST00000369519.3 | Ensembl |
| Affymetrix Human Clariom D Assay | TC1000008898.hg.1 | ENST00000369519.3 | Ensembl |
| Affymetrix Human Clariom D Assay | TC1000008898.hg.1 | ENST00000369519.3 | Ensembl |
| Affymetrix Human Clariom D Assay | TC1000008898.hg.1 | ENST00000369519.3 | Ensembl |
| Affymetrix Human Clariom D Assay | TC1000008898.hg.1 | ENST00000369519.3 | Ensembl |
| Affymetrix Human Clariom D Assay | TC1000008898.hg.1 | ENST00000465774.2 | Ensembl |
| Affymetrix Human Clariom D Assay | TC1000008898.hg.1 | ENST00000465774.2 | Ensembl |
| Affymetrix Human Clariom D Assay | TC1000008898.hg.1 | ENST00000465774.2 | Ensembl |
| Affymetrix Human Clariom D Assay | TC1000008898.hg.1 | ENST00000471172.1 | Ensembl |
| Affymetrix Human Clariom D Assay | TC1000008898.hg.1 | ENST00000471172.1 | Ensembl |
| Affymetrix Human Clariom D Assay | TC1000008898.hg.1 | ENST00000471172.1 | Ensembl |
| Affymetrix Human Clariom D Assay | TC1000008898.hg.1 | ENST00000480343.2 | Ensembl |
| Affymetrix Human Clariom D Assay | TC1000008898.hg.1 | ENST00000480343.2 | Ensembl |
| Affymetrix Human Clariom D Assay | TC1000008898.hg.1 | ENST00000480343.2 | Ensembl |
| Affymetrix Human Clariom D Assay | TC1000008898.hg.1 | ENST00000480343.2 | Ensembl |
| Affymetrix Human Clariom D Assay | TC1000008898.hg.1 | NM_001134363.2 | Refseq |
| Affymetrix Human Clariom D Assay | TC1000008898.hg.1 | NM_001134363.2 | Refseq |
| Affymetrix Human Clariom D Assay | TC1000008898.hg.1 | NM_001134363.2 | Refseq |
| Affymetrix Human Clariom D Assay | TC1000008898.hg.1 | NM_001134363.2 | Refseq |
| Affymetrix Human Clariom D Assay | TC1000008898.hg.1 | NM_001134363.2 | Refseq |
| Affymetrix Human Clariom D Assay | TC1000008898.hg.1 | NM_001134363.2 | Refseq |
| Affymetrix Human Clariom D Assay | TC1000008898.hg.1 | NM_001134363.2 | Refseq |
| Affymetrix Human Clariom D Assay | TC1000008898.hg.1 | NM_001134363.2 | Refseq |
| Affymetrix Human Clariom D Assay | TC1000008898.hg.1 | NM_001134363.2 | Refseq |
| Affymetrix Human Clariom D Assay | TC1000008898.hg.1 | NM_001134363.2 | Refseq |
| Affymetrix Human Clariom D Assay | TC1000008898.hg.1 | NM_001134363.2 | Refseq |
| Affymetrix Human Clariom D Assay | TC1000008898.hg.1 | NM_001134363.2 | Refseq |
| Affymetrix Human Clariom D Assay | TC1000008898.hg.1 | NM_001134363.2 | Refseq |
| Affymetrix Human Clariom D Assay | TC1000008898.hg.1 | NM_001134363.2 | Refseq |
| Affymetrix Human Clariom D Assay | TC1000008898.hg.1 | NM_001134363.2 | Refseq |
| Affymetrix Human Clariom D Assay | TC1000008898.hg.1 | NM_001134363.2 | Refseq |
| Affymetrix Human Clariom D Assay | TC1000008898.hg.1 | NM_001134363.2 | Refseq |
| Affymetrix Human Clariom D Assay | TC1000008898.hg.1 | NM_001134363.2 | Refseq |

RBM20
Genomic Map