Assay Details
| Gene Symbol: ZCCHC4 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | NM_024936.2 | NP_079212.2 | 9 - 10 | 1167 | 132 | |
| XM_011513835.1 | XP_011512137.1 | 10 - 11 | 1179 | 132 | ||
| XM_017008128.1 | XP_016863617.1 | 8 - 9 | 1046 | 132 | ||
| XM_017008129.1 | XP_016863618.1 | 7 - 8 | 882 | 132 | ||
| GenBank mRNA | AK026677.1 | - | 4 - 5 | 436 | 132 | |
| AK299211.1 | - | 9 - 10 | 1157 | 132 | ||
| AY629351.1 | - | 9 - 10 | 1160 | 132 | ||
| BC016914.1 | - | 6 - 7 | 821 | 132 | ||
| BC157834.1 | - | 9 - 10 | 1161 | 9057053 | 132 | |
| BC171821.1 | - | 9 - 10 | 1161 | 9054475 | 132 | |
More Information
Updated Mapping Information
This assay targets the same location as HTA transcript cluster(s) TC04000175.hg.1
Set Membership: |
|
Matching Ambion siRNAs:
Panther Classification:
Gene Ontology Categories:
MicroArray Probes:
Affymetrix
| Array Name | Probe IDs | Targets | Target Type |
|---|---|---|---|
| Affymetrix Human Transcriptome Array 2.0 | TC0400007095.hg.1 | ENST00000302874.8 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0400007095.hg.1 | ENST00000302874.8 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0400007095.hg.1 | ENST00000302874.8 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0400007095.hg.1 | ENST00000302874.8 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0400007095.hg.1 | ENST00000302874.8 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0400007095.hg.1 | ENST00000302874.8 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0400007095.hg.1 | ENST00000302874.8 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0400007095.hg.1 | ENST00000302874.8 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0400007095.hg.1 | ENST00000302874.8 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0400007095.hg.1 | ENST00000302874.8 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0400007095.hg.1 | ENST00000302874.8 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0400007095.hg.1 | ENST00000302874.8 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0400007095.hg.1 | ENST00000302874.8 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0400007095.hg.1 | ENST00000302874.8 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0400007095.hg.1 | ENST00000302874.8 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0400007095.hg.1 | ENST00000302874.8 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0400007095.hg.1 | ENST00000302874.8 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0400007095.hg.1 | ENST00000302874.8 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0400007095.hg.1 | ENST00000505412.1 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0400007095.hg.1 | ENST00000505412.1 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0400007095.hg.1 | ENST00000505412.1 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0400007095.hg.1 | ENST00000505412.1 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0400007095.hg.1 | ENST00000505412.1 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0400007095.hg.1 | ENST00000505412.1 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0400007095.hg.1 | ENST00000505412.1 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0400007095.hg.1 | ENST00000505412.1 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0400007095.hg.1 | ENST00000505412.1 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0400007095.hg.1 | ENST00000505412.1 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0400007095.hg.1 | ENST00000505412.1 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0400007095.hg.1 | ENST00000505412.1 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0400007095.hg.1 | ENST00000505451.5 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0400007095.hg.1 | ENST00000505451.5 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0400007095.hg.1 | ENST00000505451.5 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0400007095.hg.1 | ENST00000505451.5 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0400007095.hg.1 | ENST00000505451.5 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0400007095.hg.1 | ENST00000505451.5 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0400007095.hg.1 | ENST00000505451.5 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0400007095.hg.1 | ENST00000505451.5 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0400007095.hg.1 | ENST00000505451.5 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0400007095.hg.1 | ENST00000505451.5 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0400007095.hg.1 | ENST00000507760.5 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0400007095.hg.1 | ENST00000507760.5 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0400007095.hg.1 | ENST00000507760.5 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0400007095.hg.1 | ENST00000507760.5 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0400007095.hg.1 | ENST00000507760.5 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0400007095.hg.1 | ENST00000507760.5 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0400007095.hg.1 | ENST00000507760.5 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0400007095.hg.1 | ENST00000507760.5 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0400007095.hg.1 | ENST00000507760.5 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0400007095.hg.1 | ENST00000507760.5 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0400007095.hg.1 | ENST00000507760.5 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0400007095.hg.1 | ENST00000507760.5 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0400007095.hg.1 | ENST00000508058.1 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0400007095.hg.1 | ENST00000508058.1 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0400007095.hg.1 | ENST00000508058.1 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0400007095.hg.1 | ENST00000508058.1 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0400007095.hg.1 | ENST00000508058.1 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0400007095.hg.1 | ENST00000508058.1 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0400007095.hg.1 | ENST00000580712.1 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0400007095.hg.1 | ENST00000612982.1 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0400007095.hg.1 | ENST00000612982.1 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0400007095.hg.1 | ENST00000612982.1 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0400007095.hg.1 | ENST00000612982.1 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0400007095.hg.1 | ENST00000612982.1 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0400007095.hg.1 | ENST00000612982.1 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0400007095.hg.1 | ENST00000612982.1 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0400007095.hg.1 | ENST00000612982.1 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0400007095.hg.1 | ENST00000612982.1 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0400007095.hg.1 | NM_024936.2 | Refseq |
| Affymetrix Human Transcriptome Array 2.0 | TC0400007095.hg.1 | NM_024936.2 | Refseq |
| Affymetrix Human Transcriptome Array 2.0 | TC0400007095.hg.1 | NM_024936.2 | Refseq |
| Affymetrix Human Transcriptome Array 2.0 | TC0400007095.hg.1 | NM_024936.2 | Refseq |
| Affymetrix Human Transcriptome Array 2.0 | TC0400007095.hg.1 | NM_024936.2 | Refseq |
| Affymetrix Human Transcriptome Array 2.0 | TC0400007095.hg.1 | NM_024936.2 | Refseq |
| Affymetrix Human Transcriptome Array 2.0 | TC0400007095.hg.1 | NM_024936.2 | Refseq |
| Affymetrix Human Transcriptome Array 2.0 | TC0400007095.hg.1 | NM_024936.2 | Refseq |
| Affymetrix Human Transcriptome Array 2.0 | TC0400007095.hg.1 | NM_024936.2 | Refseq |
| Affymetrix Human Transcriptome Array 2.0 | TC0400007095.hg.1 | NM_024936.2 | Refseq |
| Affymetrix Human Transcriptome Array 2.0 | TC0400007095.hg.1 | NM_024936.2 | Refseq |
| Affymetrix Human Transcriptome Array 2.0 | TC0400007095.hg.1 | NM_024936.2 | Refseq |
| Affymetrix Human Transcriptome Array 2.0 | TC0400007095.hg.1 | NM_024936.2 | Refseq |
| Affymetrix Human Transcriptome Array 2.0 | TC0400007095.hg.1 | NM_024936.2 | Refseq |
| Affymetrix Human Transcriptome Array 2.0 | TC0400007095.hg.1 | NM_024936.2 | Refseq |
| Affymetrix Human Transcriptome Array 2.0 | TC0400007095.hg.1 | NM_024936.2 | Refseq |
| Affymetrix Human Transcriptome Array 2.0 | TC0400007095.hg.1 | NM_024936.2 | Refseq |
| Affymetrix Human Transcriptome Array 2.0 | TC0400007095.hg.1 | NM_024936.2 | Refseq |
| Affymetrix Human Transcriptome Array 2.0 | PSR04002701.hg.1 | TC04000175.hg.1 | HTA TC |
| Affymetrix Human Transcriptome Array 2.0 | PSR04002701.hg.1 | ENST00000302874.4 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | PSR04002701.hg.1 | NM_024936.2 | Refseq |
| Affymetrix Human Clariom D Assay | TC0400007095.hg.1 | ENST00000302874.8 | Ensembl |
| Affymetrix Human Clariom D Assay | TC0400007095.hg.1 | ENST00000302874.8 | Ensembl |
| Affymetrix Human Clariom D Assay | TC0400007095.hg.1 | ENST00000302874.8 | Ensembl |
| Affymetrix Human Clariom D Assay | TC0400007095.hg.1 | ENST00000302874.8 | Ensembl |
| Affymetrix Human Clariom D Assay | TC0400007095.hg.1 | ENST00000302874.8 | Ensembl |
| Affymetrix Human Clariom D Assay | TC0400007095.hg.1 | ENST00000302874.8 | Ensembl |
| Affymetrix Human Clariom D Assay | TC0400007095.hg.1 | ENST00000302874.8 | Ensembl |
| Affymetrix Human Clariom D Assay | TC0400007095.hg.1 | ENST00000302874.8 | Ensembl |
| Affymetrix Human Clariom D Assay | TC0400007095.hg.1 | ENST00000302874.8 | Ensembl |
| Affymetrix Human Clariom D Assay | TC0400007095.hg.1 | ENST00000302874.8 | Ensembl |
| Affymetrix Human Clariom D Assay | TC0400007095.hg.1 | ENST00000302874.8 | Ensembl |
| Affymetrix Human Clariom D Assay | TC0400007095.hg.1 | ENST00000302874.8 | Ensembl |
| Affymetrix Human Clariom D Assay | TC0400007095.hg.1 | ENST00000302874.8 | Ensembl |
| Affymetrix Human Clariom D Assay | TC0400007095.hg.1 | ENST00000302874.8 | Ensembl |
| Affymetrix Human Clariom D Assay | TC0400007095.hg.1 | ENST00000302874.8 | Ensembl |
| Affymetrix Human Clariom D Assay | TC0400007095.hg.1 | ENST00000302874.8 | Ensembl |
| Affymetrix Human Clariom D Assay | TC0400007095.hg.1 | ENST00000302874.8 | Ensembl |
| Affymetrix Human Clariom D Assay | TC0400007095.hg.1 | ENST00000302874.8 | Ensembl |
| Affymetrix Human Clariom D Assay | TC0400007095.hg.1 | ENST00000505412.1 | Ensembl |
| Affymetrix Human Clariom D Assay | TC0400007095.hg.1 | ENST00000505412.1 | Ensembl |
| Affymetrix Human Clariom D Assay | TC0400007095.hg.1 | ENST00000505412.1 | Ensembl |
| Affymetrix Human Clariom D Assay | TC0400007095.hg.1 | ENST00000505412.1 | Ensembl |
| Affymetrix Human Clariom D Assay | TC0400007095.hg.1 | ENST00000505412.1 | Ensembl |
| Affymetrix Human Clariom D Assay | TC0400007095.hg.1 | ENST00000505412.1 | Ensembl |
| Affymetrix Human Clariom D Assay | TC0400007095.hg.1 | ENST00000505412.1 | Ensembl |
| Affymetrix Human Clariom D Assay | TC0400007095.hg.1 | ENST00000505412.1 | Ensembl |
| Affymetrix Human Clariom D Assay | TC0400007095.hg.1 | ENST00000505412.1 | Ensembl |
| Affymetrix Human Clariom D Assay | TC0400007095.hg.1 | ENST00000505412.1 | Ensembl |
| Affymetrix Human Clariom D Assay | TC0400007095.hg.1 | ENST00000505412.1 | Ensembl |
| Affymetrix Human Clariom D Assay | TC0400007095.hg.1 | ENST00000505412.1 | Ensembl |
| Affymetrix Human Clariom D Assay | TC0400007095.hg.1 | ENST00000505451.5 | Ensembl |
| Affymetrix Human Clariom D Assay | TC0400007095.hg.1 | ENST00000505451.5 | Ensembl |
| Affymetrix Human Clariom D Assay | TC0400007095.hg.1 | ENST00000505451.5 | Ensembl |
| Affymetrix Human Clariom D Assay | TC0400007095.hg.1 | ENST00000505451.5 | Ensembl |
| Affymetrix Human Clariom D Assay | TC0400007095.hg.1 | ENST00000505451.5 | Ensembl |
| Affymetrix Human Clariom D Assay | TC0400007095.hg.1 | ENST00000505451.5 | Ensembl |
| Affymetrix Human Clariom D Assay | TC0400007095.hg.1 | ENST00000505451.5 | Ensembl |
| Affymetrix Human Clariom D Assay | TC0400007095.hg.1 | ENST00000505451.5 | Ensembl |
| Affymetrix Human Clariom D Assay | TC0400007095.hg.1 | ENST00000505451.5 | Ensembl |
| Affymetrix Human Clariom D Assay | TC0400007095.hg.1 | ENST00000505451.5 | Ensembl |
| Affymetrix Human Clariom D Assay | TC0400007095.hg.1 | ENST00000507760.5 | Ensembl |
| Affymetrix Human Clariom D Assay | TC0400007095.hg.1 | ENST00000507760.5 | Ensembl |
| Affymetrix Human Clariom D Assay | TC0400007095.hg.1 | ENST00000507760.5 | Ensembl |
| Affymetrix Human Clariom D Assay | TC0400007095.hg.1 | ENST00000507760.5 | Ensembl |
| Affymetrix Human Clariom D Assay | TC0400007095.hg.1 | ENST00000507760.5 | Ensembl |
| Affymetrix Human Clariom D Assay | TC0400007095.hg.1 | ENST00000507760.5 | Ensembl |
| Affymetrix Human Clariom D Assay | TC0400007095.hg.1 | ENST00000507760.5 | Ensembl |
| Affymetrix Human Clariom D Assay | TC0400007095.hg.1 | ENST00000507760.5 | Ensembl |
| Affymetrix Human Clariom D Assay | TC0400007095.hg.1 | ENST00000507760.5 | Ensembl |
| Affymetrix Human Clariom D Assay | TC0400007095.hg.1 | ENST00000507760.5 | Ensembl |
| Affymetrix Human Clariom D Assay | TC0400007095.hg.1 | ENST00000507760.5 | Ensembl |
| Affymetrix Human Clariom D Assay | TC0400007095.hg.1 | ENST00000507760.5 | Ensembl |
| Affymetrix Human Clariom D Assay | TC0400007095.hg.1 | ENST00000508058.1 | Ensembl |
| Affymetrix Human Clariom D Assay | TC0400007095.hg.1 | ENST00000508058.1 | Ensembl |
| Affymetrix Human Clariom D Assay | TC0400007095.hg.1 | ENST00000508058.1 | Ensembl |
| Affymetrix Human Clariom D Assay | TC0400007095.hg.1 | ENST00000508058.1 | Ensembl |
| Affymetrix Human Clariom D Assay | TC0400007095.hg.1 | ENST00000508058.1 | Ensembl |
| Affymetrix Human Clariom D Assay | TC0400007095.hg.1 | ENST00000508058.1 | Ensembl |
| Affymetrix Human Clariom D Assay | TC0400007095.hg.1 | ENST00000580712.1 | Ensembl |
| Affymetrix Human Clariom D Assay | TC0400007095.hg.1 | ENST00000612982.1 | Ensembl |
| Affymetrix Human Clariom D Assay | TC0400007095.hg.1 | ENST00000612982.1 | Ensembl |
| Affymetrix Human Clariom D Assay | TC0400007095.hg.1 | ENST00000612982.1 | Ensembl |
| Affymetrix Human Clariom D Assay | TC0400007095.hg.1 | ENST00000612982.1 | Ensembl |
| Affymetrix Human Clariom D Assay | TC0400007095.hg.1 | ENST00000612982.1 | Ensembl |
| Affymetrix Human Clariom D Assay | TC0400007095.hg.1 | ENST00000612982.1 | Ensembl |
| Affymetrix Human Clariom D Assay | TC0400007095.hg.1 | ENST00000612982.1 | Ensembl |
| Affymetrix Human Clariom D Assay | TC0400007095.hg.1 | ENST00000612982.1 | Ensembl |
| Affymetrix Human Clariom D Assay | TC0400007095.hg.1 | ENST00000612982.1 | Ensembl |
| Affymetrix Human Clariom D Assay | TC0400007095.hg.1 | NM_024936.2 | Refseq |
| Affymetrix Human Clariom D Assay | TC0400007095.hg.1 | NM_024936.2 | Refseq |
| Affymetrix Human Clariom D Assay | TC0400007095.hg.1 | NM_024936.2 | Refseq |
| Affymetrix Human Clariom D Assay | TC0400007095.hg.1 | NM_024936.2 | Refseq |
| Affymetrix Human Clariom D Assay | TC0400007095.hg.1 | NM_024936.2 | Refseq |
| Affymetrix Human Clariom D Assay | TC0400007095.hg.1 | NM_024936.2 | Refseq |
| Affymetrix Human Clariom D Assay | TC0400007095.hg.1 | NM_024936.2 | Refseq |
| Affymetrix Human Clariom D Assay | TC0400007095.hg.1 | NM_024936.2 | Refseq |
| Affymetrix Human Clariom D Assay | TC0400007095.hg.1 | NM_024936.2 | Refseq |
| Affymetrix Human Clariom D Assay | TC0400007095.hg.1 | NM_024936.2 | Refseq |
| Affymetrix Human Clariom D Assay | TC0400007095.hg.1 | NM_024936.2 | Refseq |
| Affymetrix Human Clariom D Assay | TC0400007095.hg.1 | NM_024936.2 | Refseq |
| Affymetrix Human Clariom D Assay | TC0400007095.hg.1 | NM_024936.2 | Refseq |
| Affymetrix Human Clariom D Assay | TC0400007095.hg.1 | NM_024936.2 | Refseq |
| Affymetrix Human Clariom D Assay | TC0400007095.hg.1 | NM_024936.2 | Refseq |
| Affymetrix Human Clariom D Assay | TC0400007095.hg.1 | NM_024936.2 | Refseq |
| Affymetrix Human Clariom D Assay | TC0400007095.hg.1 | NM_024936.2 | Refseq |
| Affymetrix Human Clariom D Assay | TC0400007095.hg.1 | NM_024936.2 | Refseq |
| Affymetrix Human Clariom D Assay | PSR0400148590.hg.1 | ENST00000302874.8 | Ensembl |
| Affymetrix Human Clariom D Assay | PSR0400148590.hg.1 | ENST00000505412.1 | Ensembl |
| Affymetrix Human Clariom D Assay | PSR0400148590.hg.1 | ENST00000507760.5 | Ensembl |
| Affymetrix Human Clariom D Assay | PSR0400148590.hg.1 | ENST00000508058.1 | Ensembl |
| Affymetrix Human Clariom D Assay | PSR0400148590.hg.1 | NM_024936.2 | Refseq |
| Affymetrix Human Clariom D Assay | PSR0400148590.hg.1 | ENST00000302874.8 | Ensembl |
| Affymetrix Human Clariom D Assay | PSR0400148590.hg.1 | ENST00000505412.1 | Ensembl |
| Affymetrix Human Clariom D Assay | PSR0400148590.hg.1 | ENST00000507760.5 | Ensembl |
| Affymetrix Human Clariom D Assay | PSR0400148590.hg.1 | ENST00000508058.1 | Ensembl |
| Affymetrix Human Clariom D Assay | PSR0400148590.hg.1 | NM_024936.2 | Refseq |
| Affymetrix Human Clariom D Assay | PSR0400148590.hg.1 | TC0400007095.hg.1 | Clariom D TC |

ZCCHC4
Genomic Map