Assay Details
| Gene Symbol: ATP9A | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | NM_006045.2 | NP_006036.1 | 19 - 20 | 2149 | 105 | |
| XM_011528481.2 | XP_011526783.1 | 19 - 20 | 2177 | 105 | ||
| GenBank mRNA | AB014511.1 | - | 14 - 15 | 1703 | 105 | |
| AK172802.1 | - | 17 - 18 | 1838 | 105 | ||
| AK299064.1 | - | 19 - 20 | 2078 | 105 | ||
| BC110591.1 | - | 15 - 16 | 1641 | 105 | ||
| BC110592.1 | - | 15 - 16 | 1641 | 105 | ||
More Information
Updated Mapping Information
This assay targets the same location as HTA transcript cluster(s) TC20000944.hg.1
Set Membership: |
|
Matching Ambion siRNAs:
Panther Classification:
Gene Ontology Categories:
MicroArray Probes:
Affymetrix
| Array Name | Probe IDs | Targets | Target Type |
|---|---|---|---|
| Affymetrix Human Transcriptome Array 2.0 | TC2000009461.hg.1 | ENST00000311637.9 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC2000009461.hg.1 | ENST00000311637.9 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC2000009461.hg.1 | ENST00000311637.9 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC2000009461.hg.1 | ENST00000311637.9 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC2000009461.hg.1 | ENST00000311637.9 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC2000009461.hg.1 | ENST00000311637.9 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC2000009461.hg.1 | ENST00000311637.9 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC2000009461.hg.1 | ENST00000311637.9 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC2000009461.hg.1 | ENST00000311637.9 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC2000009461.hg.1 | ENST00000311637.9 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC2000009461.hg.1 | ENST00000311637.9 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC2000009461.hg.1 | ENST00000311637.9 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC2000009461.hg.1 | ENST00000311637.9 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC2000009461.hg.1 | ENST00000311637.9 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC2000009461.hg.1 | ENST00000311637.9 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC2000009461.hg.1 | ENST00000311637.9 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC2000009461.hg.1 | ENST00000311637.9 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC2000009461.hg.1 | ENST00000311637.9 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC2000009461.hg.1 | ENST00000311637.9 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC2000009461.hg.1 | ENST00000311637.9 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC2000009461.hg.1 | ENST00000311637.9 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC2000009461.hg.1 | ENST00000311637.9 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC2000009461.hg.1 | ENST00000311637.9 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC2000009461.hg.1 | ENST00000311637.9 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC2000009461.hg.1 | ENST00000311637.9 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC2000009461.hg.1 | ENST00000338821.5 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC2000009461.hg.1 | ENST00000338821.5 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC2000009461.hg.1 | ENST00000338821.5 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC2000009461.hg.1 | ENST00000338821.5 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC2000009461.hg.1 | ENST00000338821.5 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC2000009461.hg.1 | ENST00000338821.5 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC2000009461.hg.1 | ENST00000338821.5 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC2000009461.hg.1 | ENST00000338821.5 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC2000009461.hg.1 | ENST00000338821.5 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC2000009461.hg.1 | ENST00000338821.5 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC2000009461.hg.1 | ENST00000338821.5 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC2000009461.hg.1 | ENST00000338821.5 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC2000009461.hg.1 | ENST00000338821.5 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC2000009461.hg.1 | ENST00000338821.5 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC2000009461.hg.1 | ENST00000338821.5 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC2000009461.hg.1 | ENST00000338821.5 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC2000009461.hg.1 | ENST00000338821.5 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC2000009461.hg.1 | ENST00000338821.5 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC2000009461.hg.1 | ENST00000338821.5 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC2000009461.hg.1 | ENST00000338821.5 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC2000009461.hg.1 | ENST00000338821.5 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC2000009461.hg.1 | ENST00000338821.5 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC2000009461.hg.1 | ENST00000338821.5 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC2000009461.hg.1 | ENST00000338821.5 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC2000009461.hg.1 | ENST00000338821.5 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC2000009461.hg.1 | ENST00000338821.5 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC2000009461.hg.1 | ENST00000338821.5 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC2000009461.hg.1 | ENST00000338821.5 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC2000009461.hg.1 | ENST00000338821.5 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC2000009461.hg.1 | ENST00000338821.5 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC2000009461.hg.1 | ENST00000338821.5 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC2000009461.hg.1 | ENST00000338821.5 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC2000009461.hg.1 | ENST00000338821.5 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC2000009461.hg.1 | ENST00000477492.1 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC2000009461.hg.1 | ENST00000477492.1 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC2000009461.hg.1 | ENST00000477492.1 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC2000009461.hg.1 | NM_006045.2 | Refseq |
| Affymetrix Human Transcriptome Array 2.0 | TC2000009461.hg.1 | NM_006045.2 | Refseq |
| Affymetrix Human Transcriptome Array 2.0 | TC2000009461.hg.1 | NM_006045.2 | Refseq |
| Affymetrix Human Transcriptome Array 2.0 | TC2000009461.hg.1 | NM_006045.2 | Refseq |
| Affymetrix Human Transcriptome Array 2.0 | TC2000009461.hg.1 | NM_006045.2 | Refseq |
| Affymetrix Human Transcriptome Array 2.0 | TC2000009461.hg.1 | NM_006045.2 | Refseq |
| Affymetrix Human Transcriptome Array 2.0 | TC2000009461.hg.1 | NM_006045.2 | Refseq |
| Affymetrix Human Transcriptome Array 2.0 | TC2000009461.hg.1 | NM_006045.2 | Refseq |
| Affymetrix Human Transcriptome Array 2.0 | TC2000009461.hg.1 | NM_006045.2 | Refseq |
| Affymetrix Human Transcriptome Array 2.0 | TC2000009461.hg.1 | NM_006045.2 | Refseq |
| Affymetrix Human Transcriptome Array 2.0 | TC2000009461.hg.1 | NM_006045.2 | Refseq |
| Affymetrix Human Transcriptome Array 2.0 | TC2000009461.hg.1 | NM_006045.2 | Refseq |
| Affymetrix Human Transcriptome Array 2.0 | TC2000009461.hg.1 | NM_006045.2 | Refseq |
| Affymetrix Human Transcriptome Array 2.0 | TC2000009461.hg.1 | NM_006045.2 | Refseq |
| Affymetrix Human Transcriptome Array 2.0 | TC2000009461.hg.1 | NM_006045.2 | Refseq |
| Affymetrix Human Transcriptome Array 2.0 | TC2000009461.hg.1 | NM_006045.2 | Refseq |
| Affymetrix Human Transcriptome Array 2.0 | TC2000009461.hg.1 | NM_006045.2 | Refseq |
| Affymetrix Human Transcriptome Array 2.0 | TC2000009461.hg.1 | NM_006045.2 | Refseq |
| Affymetrix Human Transcriptome Array 2.0 | TC2000009461.hg.1 | NM_006045.2 | Refseq |
| Affymetrix Human Transcriptome Array 2.0 | TC2000009461.hg.1 | NM_006045.2 | Refseq |
| Affymetrix Human Transcriptome Array 2.0 | TC2000009461.hg.1 | NM_006045.2 | Refseq |
| Affymetrix Human Transcriptome Array 2.0 | TC2000009461.hg.1 | NM_006045.2 | Refseq |
| Affymetrix Human Transcriptome Array 2.0 | TC2000009461.hg.1 | NM_006045.2 | Refseq |
| Affymetrix Human Transcriptome Array 2.0 | TC2000009461.hg.1 | NM_006045.2 | Refseq |
| Affymetrix Human Transcriptome Array 2.0 | TC2000009461.hg.1 | NM_006045.2 | Refseq |
| Affymetrix Human Transcriptome Array 2.0 | TC2000009461.hg.1 | NM_006045.2 | Refseq |
| Affymetrix Human Transcriptome Array 2.0 | TC2000009461.hg.1 | NM_006045.2 | Refseq |
| Affymetrix Human Transcriptome Array 2.0 | TC2000009461.hg.1 | NM_006045.2 | Refseq |
| Affymetrix Human Transcriptome Array 2.0 | TC2000009461.hg.1 | NM_006045.2 | Refseq |
| Affymetrix Human Transcriptome Array 2.0 | TC2000009461.hg.1 | NM_006045.2 | Refseq |
| Affymetrix Human Transcriptome Array 2.0 | TC2000009461.hg.1 | NM_006045.2 | Refseq |
| Affymetrix Human Transcriptome Array 2.0 | PSR20013026.hg.1 | ENST00000311637.5 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | PSR20013026.hg.1 | ENST00000338821.5 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | PSR20013026.hg.1 | ENST00000402822.1 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | PSR20013026.hg.1 | NM_006045.2 | Refseq |
| Affymetrix Human Transcriptome Array 2.0 | PSR20013026.hg.1 | TC20000944.hg.1 | HTA TC |
| Affymetrix Human Clariom D Assay | TC2000009461.hg.1 | ENST00000311637.9 | Ensembl |
| Affymetrix Human Clariom D Assay | TC2000009461.hg.1 | ENST00000311637.9 | Ensembl |
| Affymetrix Human Clariom D Assay | TC2000009461.hg.1 | ENST00000311637.9 | Ensembl |
| Affymetrix Human Clariom D Assay | TC2000009461.hg.1 | ENST00000311637.9 | Ensembl |
| Affymetrix Human Clariom D Assay | TC2000009461.hg.1 | ENST00000311637.9 | Ensembl |
| Affymetrix Human Clariom D Assay | TC2000009461.hg.1 | ENST00000311637.9 | Ensembl |
| Affymetrix Human Clariom D Assay | TC2000009461.hg.1 | ENST00000311637.9 | Ensembl |
| Affymetrix Human Clariom D Assay | TC2000009461.hg.1 | ENST00000311637.9 | Ensembl |
| Affymetrix Human Clariom D Assay | TC2000009461.hg.1 | ENST00000311637.9 | Ensembl |
| Affymetrix Human Clariom D Assay | TC2000009461.hg.1 | ENST00000311637.9 | Ensembl |
| Affymetrix Human Clariom D Assay | TC2000009461.hg.1 | ENST00000311637.9 | Ensembl |
| Affymetrix Human Clariom D Assay | TC2000009461.hg.1 | ENST00000311637.9 | Ensembl |
| Affymetrix Human Clariom D Assay | TC2000009461.hg.1 | ENST00000311637.9 | Ensembl |
| Affymetrix Human Clariom D Assay | TC2000009461.hg.1 | ENST00000311637.9 | Ensembl |
| Affymetrix Human Clariom D Assay | TC2000009461.hg.1 | ENST00000311637.9 | Ensembl |
| Affymetrix Human Clariom D Assay | TC2000009461.hg.1 | ENST00000311637.9 | Ensembl |
| Affymetrix Human Clariom D Assay | TC2000009461.hg.1 | ENST00000311637.9 | Ensembl |
| Affymetrix Human Clariom D Assay | TC2000009461.hg.1 | ENST00000311637.9 | Ensembl |
| Affymetrix Human Clariom D Assay | TC2000009461.hg.1 | ENST00000311637.9 | Ensembl |
| Affymetrix Human Clariom D Assay | TC2000009461.hg.1 | ENST00000311637.9 | Ensembl |
| Affymetrix Human Clariom D Assay | TC2000009461.hg.1 | ENST00000311637.9 | Ensembl |
| Affymetrix Human Clariom D Assay | TC2000009461.hg.1 | ENST00000311637.9 | Ensembl |
| Affymetrix Human Clariom D Assay | TC2000009461.hg.1 | ENST00000311637.9 | Ensembl |
| Affymetrix Human Clariom D Assay | TC2000009461.hg.1 | ENST00000311637.9 | Ensembl |
| Affymetrix Human Clariom D Assay | TC2000009461.hg.1 | ENST00000311637.9 | Ensembl |
| Affymetrix Human Clariom D Assay | TC2000009461.hg.1 | ENST00000338821.5 | Ensembl |
| Affymetrix Human Clariom D Assay | TC2000009461.hg.1 | ENST00000338821.5 | Ensembl |
| Affymetrix Human Clariom D Assay | TC2000009461.hg.1 | ENST00000338821.5 | Ensembl |
| Affymetrix Human Clariom D Assay | TC2000009461.hg.1 | ENST00000338821.5 | Ensembl |
| Affymetrix Human Clariom D Assay | TC2000009461.hg.1 | ENST00000338821.5 | Ensembl |
| Affymetrix Human Clariom D Assay | TC2000009461.hg.1 | ENST00000338821.5 | Ensembl |
| Affymetrix Human Clariom D Assay | TC2000009461.hg.1 | ENST00000338821.5 | Ensembl |
| Affymetrix Human Clariom D Assay | TC2000009461.hg.1 | ENST00000338821.5 | Ensembl |
| Affymetrix Human Clariom D Assay | TC2000009461.hg.1 | ENST00000338821.5 | Ensembl |
| Affymetrix Human Clariom D Assay | TC2000009461.hg.1 | ENST00000338821.5 | Ensembl |
| Affymetrix Human Clariom D Assay | TC2000009461.hg.1 | ENST00000338821.5 | Ensembl |
| Affymetrix Human Clariom D Assay | TC2000009461.hg.1 | ENST00000338821.5 | Ensembl |
| Affymetrix Human Clariom D Assay | TC2000009461.hg.1 | ENST00000338821.5 | Ensembl |
| Affymetrix Human Clariom D Assay | TC2000009461.hg.1 | ENST00000338821.5 | Ensembl |
| Affymetrix Human Clariom D Assay | TC2000009461.hg.1 | ENST00000338821.5 | Ensembl |
| Affymetrix Human Clariom D Assay | TC2000009461.hg.1 | ENST00000338821.5 | Ensembl |
| Affymetrix Human Clariom D Assay | TC2000009461.hg.1 | ENST00000338821.5 | Ensembl |
| Affymetrix Human Clariom D Assay | TC2000009461.hg.1 | ENST00000338821.5 | Ensembl |
| Affymetrix Human Clariom D Assay | TC2000009461.hg.1 | ENST00000338821.5 | Ensembl |
| Affymetrix Human Clariom D Assay | TC2000009461.hg.1 | ENST00000338821.5 | Ensembl |
| Affymetrix Human Clariom D Assay | TC2000009461.hg.1 | ENST00000338821.5 | Ensembl |
| Affymetrix Human Clariom D Assay | TC2000009461.hg.1 | ENST00000338821.5 | Ensembl |
| Affymetrix Human Clariom D Assay | TC2000009461.hg.1 | ENST00000338821.5 | Ensembl |
| Affymetrix Human Clariom D Assay | TC2000009461.hg.1 | ENST00000338821.5 | Ensembl |
| Affymetrix Human Clariom D Assay | TC2000009461.hg.1 | ENST00000338821.5 | Ensembl |
| Affymetrix Human Clariom D Assay | TC2000009461.hg.1 | ENST00000338821.5 | Ensembl |
| Affymetrix Human Clariom D Assay | TC2000009461.hg.1 | ENST00000338821.5 | Ensembl |
| Affymetrix Human Clariom D Assay | TC2000009461.hg.1 | ENST00000338821.5 | Ensembl |
| Affymetrix Human Clariom D Assay | TC2000009461.hg.1 | ENST00000338821.5 | Ensembl |
| Affymetrix Human Clariom D Assay | TC2000009461.hg.1 | ENST00000338821.5 | Ensembl |
| Affymetrix Human Clariom D Assay | TC2000009461.hg.1 | ENST00000338821.5 | Ensembl |
| Affymetrix Human Clariom D Assay | TC2000009461.hg.1 | ENST00000338821.5 | Ensembl |
| Affymetrix Human Clariom D Assay | TC2000009461.hg.1 | ENST00000338821.5 | Ensembl |
| Affymetrix Human Clariom D Assay | TC2000009461.hg.1 | ENST00000477492.1 | Ensembl |
| Affymetrix Human Clariom D Assay | TC2000009461.hg.1 | ENST00000477492.1 | Ensembl |
| Affymetrix Human Clariom D Assay | TC2000009461.hg.1 | ENST00000477492.1 | Ensembl |
| Affymetrix Human Clariom D Assay | TC2000009461.hg.1 | NM_006045.2 | Refseq |
| Affymetrix Human Clariom D Assay | TC2000009461.hg.1 | NM_006045.2 | Refseq |
| Affymetrix Human Clariom D Assay | TC2000009461.hg.1 | NM_006045.2 | Refseq |
| Affymetrix Human Clariom D Assay | TC2000009461.hg.1 | NM_006045.2 | Refseq |
| Affymetrix Human Clariom D Assay | TC2000009461.hg.1 | NM_006045.2 | Refseq |
| Affymetrix Human Clariom D Assay | TC2000009461.hg.1 | NM_006045.2 | Refseq |
| Affymetrix Human Clariom D Assay | TC2000009461.hg.1 | NM_006045.2 | Refseq |
| Affymetrix Human Clariom D Assay | TC2000009461.hg.1 | NM_006045.2 | Refseq |
| Affymetrix Human Clariom D Assay | TC2000009461.hg.1 | NM_006045.2 | Refseq |
| Affymetrix Human Clariom D Assay | TC2000009461.hg.1 | NM_006045.2 | Refseq |
| Affymetrix Human Clariom D Assay | TC2000009461.hg.1 | NM_006045.2 | Refseq |
| Affymetrix Human Clariom D Assay | TC2000009461.hg.1 | NM_006045.2 | Refseq |
| Affymetrix Human Clariom D Assay | TC2000009461.hg.1 | NM_006045.2 | Refseq |
| Affymetrix Human Clariom D Assay | TC2000009461.hg.1 | NM_006045.2 | Refseq |
| Affymetrix Human Clariom D Assay | TC2000009461.hg.1 | NM_006045.2 | Refseq |
| Affymetrix Human Clariom D Assay | TC2000009461.hg.1 | NM_006045.2 | Refseq |
| Affymetrix Human Clariom D Assay | TC2000009461.hg.1 | NM_006045.2 | Refseq |
| Affymetrix Human Clariom D Assay | TC2000009461.hg.1 | NM_006045.2 | Refseq |
| Affymetrix Human Clariom D Assay | TC2000009461.hg.1 | NM_006045.2 | Refseq |
| Affymetrix Human Clariom D Assay | TC2000009461.hg.1 | NM_006045.2 | Refseq |
| Affymetrix Human Clariom D Assay | TC2000009461.hg.1 | NM_006045.2 | Refseq |
| Affymetrix Human Clariom D Assay | TC2000009461.hg.1 | NM_006045.2 | Refseq |
| Affymetrix Human Clariom D Assay | TC2000009461.hg.1 | NM_006045.2 | Refseq |
| Affymetrix Human Clariom D Assay | TC2000009461.hg.1 | NM_006045.2 | Refseq |
| Affymetrix Human Clariom D Assay | TC2000009461.hg.1 | NM_006045.2 | Refseq |
| Affymetrix Human Clariom D Assay | TC2000009461.hg.1 | NM_006045.2 | Refseq |
| Affymetrix Human Clariom D Assay | TC2000009461.hg.1 | NM_006045.2 | Refseq |
| Affymetrix Human Clariom D Assay | TC2000009461.hg.1 | NM_006045.2 | Refseq |
| Affymetrix Human Clariom D Assay | TC2000009461.hg.1 | NM_006045.2 | Refseq |
| Affymetrix Human Clariom D Assay | TC2000009461.hg.1 | NM_006045.2 | Refseq |
| Affymetrix Human Clariom D Assay | TC2000009461.hg.1 | NM_006045.2 | Refseq |
| Affymetrix Human Clariom D Assay | PSR2000166161.hg.1 | ENST00000311637.9 | Ensembl |
| Affymetrix Human Clariom D Assay | PSR2000166161.hg.1 | ENST00000338821.5 | Ensembl |
| Affymetrix Human Clariom D Assay | PSR2000166161.hg.1 | NM_006045.2 | Refseq |
| Affymetrix Human Clariom D Assay | PSR2000166161.hg.1 | TC2000009461.hg.1 | Clariom D TC |
| Affymetrix Human Clariom D Assay | PSR2000166161.hg.1 | ENST00000311637.9 | Ensembl |
| Affymetrix Human Clariom D Assay | PSR2000166161.hg.1 | ENST00000338821.5 | Ensembl |
| Affymetrix Human Clariom D Assay | PSR2000166161.hg.1 | NM_006045.2 | Refseq |

ATP9A
Genomic Map