Assay Details
| Gene Symbol: C16orf52 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| GenBank mRNA | BC027604.1 | - | 3 - 4 | 518 | 172 | |
| BC053950.1 | - | 3 - 4 | 505 | 172 | ||
Adjacent Gene Details
| Gene name | Gene Symbol | Alias | Distance | Relative Direction | Location Chromsome | |
|---|---|---|---|---|---|---|
| Upstream | PDZ domain containing 9 | PDZD9 | C16orf65 | -74.4kb | Anti-sense | Chr. 16 - 21957540 - 22001137 on NCBI Build GRCh38 |
| Downstream | von Willebrand factor A domain containing 3A | VWA3A | 8.0kb | Sense | Chr. 16 - 22091930 - 22156966 on NCBI Build GRCh38 |
More Information
Set Membership: |
|
MicroArray Probes:
Affymetrix
| Array Name | Probe IDs | Targets | Target Type |
|---|---|---|---|
| Affymetrix Human Transcriptome Array 2.0 | TC1600007186.hg.1 | ENST00000331057.8 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC1600007186.hg.1 | ENST00000331057.8 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC1600007186.hg.1 | ENST00000331057.8 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC1600007186.hg.1 | ENST00000331057.8 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC1600007186.hg.1 | ENST00000331057.8 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC1600007186.hg.1 | ENST00000331057.8 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC1600007186.hg.1 | ENST00000331057.8 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC1600007186.hg.1 | ENST00000331057.8 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC1600007186.hg.1 | ENST00000542527.6 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC1600007186.hg.1 | ENST00000542527.6 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC1600007186.hg.1 | ENST00000542527.6 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC1600007186.hg.1 | ENST00000542527.6 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC1600007186.hg.1 | ENST00000542527.6 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC1600007186.hg.1 | ENST00000542527.6 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC1600007186.hg.1 | ENST00000542527.6 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC1600007186.hg.1 | ENST00000542527.6 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC1600007186.hg.1 | ENST00000542527.6 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC1600007186.hg.1 | ENST00000542527.6 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC1600007186.hg.1 | ENST00000562186.1 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC1600007186.hg.1 | ENST00000562186.1 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC1600007186.hg.1 | ENST00000562186.1 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC1600007186.hg.1 | ENST00000562186.1 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC1600007186.hg.1 | ENST00000562695.1 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC1600007186.hg.1 | ENST00000562695.1 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC1600007186.hg.1 | ENST00000562695.1 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC1600007186.hg.1 | ENST00000567004.1 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC1600007186.hg.1 | ENST00000567004.1 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC1600007186.hg.1 | ENST00000567004.1 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC1600007186.hg.1 | ENST00000567004.1 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC1600007186.hg.1 | ENST00000568049.1 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC1600007186.hg.1 | ENST00000568049.1 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC1600007186.hg.1 | ENST00000568049.1 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC1600007186.hg.1 | ENST00000568049.1 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC1600007186.hg.1 | ENST00000569656.5 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC1600007186.hg.1 | ENST00000569656.5 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC1600007186.hg.1 | ENST00000569656.5 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC1600007186.hg.1 | ENST00000569656.5 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC1600007186.hg.1 | ENST00000569656.5 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC1600007186.hg.1 | ENST00000569656.5 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC1600007186.hg.1 | ENST00000569656.5 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC1600007186.hg.1 | NM_001164579.1 | Refseq |
| Affymetrix Human Transcriptome Array 2.0 | TC1600007186.hg.1 | NM_001164579.1 | Refseq |
| Affymetrix Human Transcriptome Array 2.0 | TC1600007186.hg.1 | NM_001164579.1 | Refseq |
| Affymetrix Human Transcriptome Array 2.0 | TC1600007186.hg.1 | NM_001164579.1 | Refseq |
| Affymetrix Human Transcriptome Array 2.0 | TC1600007186.hg.1 | NM_001164579.1 | Refseq |
| Affymetrix Human Transcriptome Array 2.0 | TC1600007186.hg.1 | NM_001164579.1 | Refseq |
| Affymetrix Human Transcriptome Array 2.0 | TC1600007186.hg.1 | NM_001164579.1 | Refseq |
| Affymetrix Human Transcriptome Array 2.0 | TC1600007186.hg.1 | NM_001164579.1 | Refseq |
| Affymetrix Human Transcriptome Array 2.0 | TC1600007186.hg.1 | NM_001164579.1 | Refseq |
| Affymetrix Human Transcriptome Array 2.0 | TC1600007186.hg.1 | NM_001164579.1 | Refseq |
| Affymetrix Human Transcriptome Array 2.0 | TC1600007186.hg.1 | NM_001164579.1 | Refseq |
| Affymetrix Human Clariom D Assay | TC1600007186.hg.1 | ENST00000331057.8 | Ensembl |
| Affymetrix Human Clariom D Assay | TC1600007186.hg.1 | ENST00000331057.8 | Ensembl |
| Affymetrix Human Clariom D Assay | TC1600007186.hg.1 | ENST00000331057.8 | Ensembl |
| Affymetrix Human Clariom D Assay | TC1600007186.hg.1 | ENST00000331057.8 | Ensembl |
| Affymetrix Human Clariom D Assay | TC1600007186.hg.1 | ENST00000331057.8 | Ensembl |
| Affymetrix Human Clariom D Assay | TC1600007186.hg.1 | ENST00000331057.8 | Ensembl |
| Affymetrix Human Clariom D Assay | TC1600007186.hg.1 | ENST00000331057.8 | Ensembl |
| Affymetrix Human Clariom D Assay | TC1600007186.hg.1 | ENST00000331057.8 | Ensembl |
| Affymetrix Human Clariom D Assay | TC1600007186.hg.1 | ENST00000542527.6 | Ensembl |
| Affymetrix Human Clariom D Assay | TC1600007186.hg.1 | ENST00000542527.6 | Ensembl |
| Affymetrix Human Clariom D Assay | TC1600007186.hg.1 | ENST00000542527.6 | Ensembl |
| Affymetrix Human Clariom D Assay | TC1600007186.hg.1 | ENST00000542527.6 | Ensembl |
| Affymetrix Human Clariom D Assay | TC1600007186.hg.1 | ENST00000542527.6 | Ensembl |
| Affymetrix Human Clariom D Assay | TC1600007186.hg.1 | ENST00000542527.6 | Ensembl |
| Affymetrix Human Clariom D Assay | TC1600007186.hg.1 | ENST00000542527.6 | Ensembl |
| Affymetrix Human Clariom D Assay | TC1600007186.hg.1 | ENST00000542527.6 | Ensembl |
| Affymetrix Human Clariom D Assay | TC1600007186.hg.1 | ENST00000542527.6 | Ensembl |
| Affymetrix Human Clariom D Assay | TC1600007186.hg.1 | ENST00000542527.6 | Ensembl |
| Affymetrix Human Clariom D Assay | TC1600007186.hg.1 | ENST00000562186.1 | Ensembl |
| Affymetrix Human Clariom D Assay | TC1600007186.hg.1 | ENST00000562186.1 | Ensembl |
| Affymetrix Human Clariom D Assay | TC1600007186.hg.1 | ENST00000562186.1 | Ensembl |
| Affymetrix Human Clariom D Assay | TC1600007186.hg.1 | ENST00000562186.1 | Ensembl |
| Affymetrix Human Clariom D Assay | TC1600007186.hg.1 | ENST00000562695.1 | Ensembl |
| Affymetrix Human Clariom D Assay | TC1600007186.hg.1 | ENST00000562695.1 | Ensembl |
| Affymetrix Human Clariom D Assay | TC1600007186.hg.1 | ENST00000562695.1 | Ensembl |
| Affymetrix Human Clariom D Assay | TC1600007186.hg.1 | ENST00000567004.1 | Ensembl |
| Affymetrix Human Clariom D Assay | TC1600007186.hg.1 | ENST00000567004.1 | Ensembl |
| Affymetrix Human Clariom D Assay | TC1600007186.hg.1 | ENST00000567004.1 | Ensembl |
| Affymetrix Human Clariom D Assay | TC1600007186.hg.1 | ENST00000567004.1 | Ensembl |
| Affymetrix Human Clariom D Assay | TC1600007186.hg.1 | ENST00000568049.1 | Ensembl |
| Affymetrix Human Clariom D Assay | TC1600007186.hg.1 | ENST00000568049.1 | Ensembl |
| Affymetrix Human Clariom D Assay | TC1600007186.hg.1 | ENST00000568049.1 | Ensembl |
| Affymetrix Human Clariom D Assay | TC1600007186.hg.1 | ENST00000568049.1 | Ensembl |
| Affymetrix Human Clariom D Assay | TC1600007186.hg.1 | ENST00000569656.5 | Ensembl |
| Affymetrix Human Clariom D Assay | TC1600007186.hg.1 | ENST00000569656.5 | Ensembl |
| Affymetrix Human Clariom D Assay | TC1600007186.hg.1 | ENST00000569656.5 | Ensembl |
| Affymetrix Human Clariom D Assay | TC1600007186.hg.1 | ENST00000569656.5 | Ensembl |
| Affymetrix Human Clariom D Assay | TC1600007186.hg.1 | ENST00000569656.5 | Ensembl |
| Affymetrix Human Clariom D Assay | TC1600007186.hg.1 | ENST00000569656.5 | Ensembl |
| Affymetrix Human Clariom D Assay | TC1600007186.hg.1 | ENST00000569656.5 | Ensembl |
| Affymetrix Human Clariom D Assay | TC1600007186.hg.1 | NM_001164579.1 | Refseq |
| Affymetrix Human Clariom D Assay | TC1600007186.hg.1 | NM_001164579.1 | Refseq |
| Affymetrix Human Clariom D Assay | TC1600007186.hg.1 | NM_001164579.1 | Refseq |
| Affymetrix Human Clariom D Assay | TC1600007186.hg.1 | NM_001164579.1 | Refseq |
| Affymetrix Human Clariom D Assay | TC1600007186.hg.1 | NM_001164579.1 | Refseq |
| Affymetrix Human Clariom D Assay | TC1600007186.hg.1 | NM_001164579.1 | Refseq |
| Affymetrix Human Clariom D Assay | TC1600007186.hg.1 | NM_001164579.1 | Refseq |
| Affymetrix Human Clariom D Assay | TC1600007186.hg.1 | NM_001164579.1 | Refseq |
| Affymetrix Human Clariom D Assay | TC1600007186.hg.1 | NM_001164579.1 | Refseq |
| Affymetrix Human Clariom D Assay | TC1600007186.hg.1 | NM_001164579.1 | Refseq |
| Affymetrix Human Clariom D Assay | TC1600007186.hg.1 | NM_001164579.1 | Refseq |

C16orf52
Genomic Map