Assay Details
| Gene Symbol: DLGAP2 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | NM_004745.4 | NP_004736.2 | 6 - 7 | 2051 | 56 | |
| GenBank mRNA | AB000275.1 | - | 3 - 4 | 606 | 56 | |
| AF119817.1 | - | 6 - 7 | 2036 | 56 | ||
| AF119818.1 | - | 6 - 7 | 2051 | 56 | ||
| BC141862.1 | - | 5 - 6 | 1967 | 40027523 | 56 | |
| KJ534823.1 | - | 5 - 6 | 1917 | 56 | ||
More Information
Updated Mapping Information
This assay targets the same location as HTA transcript cluster(s) TC08000007.hg.1
Set Membership: |
|
Matching Ambion siRNAs:
Panther Classification:
Gene Ontology Categories:
MicroArray Probes:
Affymetrix
| Array Name | Probe IDs | Targets | Target Type |
|---|---|---|---|
| Affymetrix Human Transcriptome Array 2.0 | PSR08000108.hg.1 | TC08000007.hg.1 | HTA TC |
| Affymetrix Human Transcriptome Array 2.0 | PSR08000108.hg.1 | ENST00000356067.4 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | PSR08000108.hg.1 | ENST00000421627.2 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | PSR08000108.hg.1 | NM_004745.4 | Refseq |
| Affymetrix Human Transcriptome Array 2.0 | PSR08000106.hg.1 | TC08000007.hg.1 | HTA TC |
| Affymetrix Human Transcriptome Array 2.0 | PSR08000106.hg.1 | ENST00000421627.2 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | PSR08000106.hg.1 | NM_004745.4 | Refseq |
| Affymetrix Human Transcriptome Array 2.0 | TC0800006475.hg.1 | ENST00000421627.6 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0800006475.hg.1 | ENST00000421627.6 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0800006475.hg.1 | ENST00000421627.6 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0800006475.hg.1 | ENST00000421627.6 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0800006475.hg.1 | ENST00000421627.6 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0800006475.hg.1 | ENST00000421627.6 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0800006475.hg.1 | ENST00000421627.6 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0800006475.hg.1 | ENST00000421627.6 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0800006475.hg.1 | ENST00000421627.6 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0800006475.hg.1 | ENST00000421627.6 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0800006475.hg.1 | ENST00000421627.6 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0800006475.hg.1 | ENST00000421627.6 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0800006475.hg.1 | ENST00000421627.6 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0800006475.hg.1 | ENST00000421627.6 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0800006475.hg.1 | ENST00000421627.6 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0800006475.hg.1 | ENST00000421627.6 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0800006475.hg.1 | ENST00000421627.6 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0800006475.hg.1 | ENST00000421627.6 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0800006475.hg.1 | ENST00000518530.2 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0800006475.hg.1 | ENST00000518530.2 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0800006475.hg.1 | ENST00000518530.2 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0800006475.hg.1 | ENST00000518530.2 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0800006475.hg.1 | ENST00000520901.5 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0800006475.hg.1 | ENST00000520901.5 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0800006475.hg.1 | ENST00000520901.5 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0800006475.hg.1 | ENST00000520901.5 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0800006475.hg.1 | ENST00000520901.5 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0800006475.hg.1 | ENST00000520901.5 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0800006475.hg.1 | ENST00000520901.5 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0800006475.hg.1 | ENST00000520901.5 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0800006475.hg.1 | ENST00000520901.5 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0800006475.hg.1 | ENST00000520901.5 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0800006475.hg.1 | ENST00000520901.5 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0800006475.hg.1 | ENST00000520901.5 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0800006475.hg.1 | ENST00000520901.5 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0800006475.hg.1 | ENST00000520901.5 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0800006475.hg.1 | ENST00000522499.1 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0800006475.hg.1 | ENST00000522499.1 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0800006475.hg.1 | ENST00000522499.1 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0800006475.hg.1 | ENST00000522499.1 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0800006475.hg.1 | ENST00000522499.1 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0800006475.hg.1 | ENST00000524065.4 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0800006475.hg.1 | ENST00000524065.4 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0800006475.hg.1 | ENST00000524065.4 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0800006475.hg.1 | ENST00000524065.4 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0800006475.hg.1 | ENST00000524065.4 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0800006475.hg.1 | ENST00000612087.1 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0800006475.hg.1 | ENST00000612087.1 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0800006475.hg.1 | ENST00000612087.1 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0800006475.hg.1 | ENST00000612087.1 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0800006475.hg.1 | ENST00000612087.1 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0800006475.hg.1 | ENST00000612087.1 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0800006475.hg.1 | ENST00000612087.1 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0800006475.hg.1 | ENST00000612087.1 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0800006475.hg.1 | ENST00000612087.1 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0800006475.hg.1 | ENST00000612087.1 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0800006475.hg.1 | ENST00000612087.1 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0800006475.hg.1 | ENST00000612087.1 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0800006475.hg.1 | ENST00000612087.1 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0800006475.hg.1 | ENST00000612087.1 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0800006475.hg.1 | ENST00000612087.1 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0800006475.hg.1 | ENST00000612087.1 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0800006475.hg.1 | ENST00000612087.1 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0800006475.hg.1 | NM_001277161.1 | Refseq |
| Affymetrix Human Transcriptome Array 2.0 | TC0800006475.hg.1 | NM_001277161.1 | Refseq |
| Affymetrix Human Transcriptome Array 2.0 | TC0800006475.hg.1 | NM_001277161.1 | Refseq |
| Affymetrix Human Transcriptome Array 2.0 | TC0800006475.hg.1 | NM_001277161.1 | Refseq |
| Affymetrix Human Transcriptome Array 2.0 | TC0800006475.hg.1 | NM_001277161.1 | Refseq |
| Affymetrix Human Transcriptome Array 2.0 | TC0800006475.hg.1 | NM_001277161.1 | Refseq |
| Affymetrix Human Transcriptome Array 2.0 | TC0800006475.hg.1 | NM_001277161.1 | Refseq |
| Affymetrix Human Transcriptome Array 2.0 | TC0800006475.hg.1 | NM_001277161.1 | Refseq |
| Affymetrix Human Transcriptome Array 2.0 | TC0800006475.hg.1 | NM_001277161.1 | Refseq |
| Affymetrix Human Transcriptome Array 2.0 | TC0800006475.hg.1 | NM_001277161.1 | Refseq |
| Affymetrix Human Transcriptome Array 2.0 | TC0800006475.hg.1 | NM_001277161.1 | Refseq |
| Affymetrix Human Transcriptome Array 2.0 | TC0800006475.hg.1 | NM_001277161.1 | Refseq |
| Affymetrix Human Transcriptome Array 2.0 | TC0800006475.hg.1 | NM_001277161.1 | Refseq |
| Affymetrix Human Transcriptome Array 2.0 | TC0800006475.hg.1 | NM_001277161.1 | Refseq |
| Affymetrix Human Transcriptome Array 2.0 | TC0800006475.hg.1 | NM_001277161.1 | Refseq |
| Affymetrix Human Transcriptome Array 2.0 | TC0800006475.hg.1 | NM_001277161.1 | Refseq |
| Affymetrix Human Transcriptome Array 2.0 | TC0800006475.hg.1 | NM_001277161.1 | Refseq |
| Affymetrix Human Transcriptome Array 2.0 | TC0800006475.hg.1 | NM_004745.4 | Refseq |
| Affymetrix Human Transcriptome Array 2.0 | TC0800006475.hg.1 | NM_004745.4 | Refseq |
| Affymetrix Human Transcriptome Array 2.0 | TC0800006475.hg.1 | NM_004745.4 | Refseq |
| Affymetrix Human Transcriptome Array 2.0 | TC0800006475.hg.1 | NM_004745.4 | Refseq |
| Affymetrix Human Transcriptome Array 2.0 | TC0800006475.hg.1 | NM_004745.4 | Refseq |
| Affymetrix Human Transcriptome Array 2.0 | TC0800006475.hg.1 | NM_004745.4 | Refseq |
| Affymetrix Human Transcriptome Array 2.0 | TC0800006475.hg.1 | NM_004745.4 | Refseq |
| Affymetrix Human Transcriptome Array 2.0 | TC0800006475.hg.1 | NM_004745.4 | Refseq |
| Affymetrix Human Transcriptome Array 2.0 | TC0800006475.hg.1 | NM_004745.4 | Refseq |
| Affymetrix Human Transcriptome Array 2.0 | TC0800006475.hg.1 | NM_004745.4 | Refseq |
| Affymetrix Human Transcriptome Array 2.0 | TC0800006475.hg.1 | NM_004745.4 | Refseq |
| Affymetrix Human Transcriptome Array 2.0 | TC0800006475.hg.1 | NM_004745.4 | Refseq |
| Affymetrix Human Transcriptome Array 2.0 | TC0800006475.hg.1 | NM_004745.4 | Refseq |
| Affymetrix Human Transcriptome Array 2.0 | TC0800006475.hg.1 | NM_004745.4 | Refseq |
| Affymetrix Human Transcriptome Array 2.0 | TC0800006475.hg.1 | NM_004745.4 | Refseq |
| Affymetrix Human Transcriptome Array 2.0 | TC0800006475.hg.1 | NM_004745.4 | Refseq |
| Affymetrix Human Transcriptome Array 2.0 | TC0800006475.hg.1 | NM_004745.4 | Refseq |
| Affymetrix Human Transcriptome Array 2.0 | TC0800006475.hg.1 | NM_004745.4 | Refseq |
| Affymetrix Human Clariom D Assay | TC0800006475.hg.1 | ENST00000421627.6 | Ensembl |
| Affymetrix Human Clariom D Assay | TC0800006475.hg.1 | ENST00000421627.6 | Ensembl |
| Affymetrix Human Clariom D Assay | TC0800006475.hg.1 | ENST00000421627.6 | Ensembl |
| Affymetrix Human Clariom D Assay | TC0800006475.hg.1 | ENST00000421627.6 | Ensembl |
| Affymetrix Human Clariom D Assay | TC0800006475.hg.1 | ENST00000421627.6 | Ensembl |
| Affymetrix Human Clariom D Assay | TC0800006475.hg.1 | ENST00000421627.6 | Ensembl |
| Affymetrix Human Clariom D Assay | TC0800006475.hg.1 | ENST00000421627.6 | Ensembl |
| Affymetrix Human Clariom D Assay | TC0800006475.hg.1 | ENST00000421627.6 | Ensembl |
| Affymetrix Human Clariom D Assay | TC0800006475.hg.1 | ENST00000421627.6 | Ensembl |
| Affymetrix Human Clariom D Assay | TC0800006475.hg.1 | ENST00000421627.6 | Ensembl |
| Affymetrix Human Clariom D Assay | TC0800006475.hg.1 | ENST00000421627.6 | Ensembl |
| Affymetrix Human Clariom D Assay | TC0800006475.hg.1 | ENST00000421627.6 | Ensembl |
| Affymetrix Human Clariom D Assay | TC0800006475.hg.1 | ENST00000421627.6 | Ensembl |
| Affymetrix Human Clariom D Assay | TC0800006475.hg.1 | ENST00000421627.6 | Ensembl |
| Affymetrix Human Clariom D Assay | TC0800006475.hg.1 | ENST00000421627.6 | Ensembl |
| Affymetrix Human Clariom D Assay | TC0800006475.hg.1 | ENST00000421627.6 | Ensembl |
| Affymetrix Human Clariom D Assay | TC0800006475.hg.1 | ENST00000421627.6 | Ensembl |
| Affymetrix Human Clariom D Assay | TC0800006475.hg.1 | ENST00000421627.6 | Ensembl |
| Affymetrix Human Clariom D Assay | TC0800006475.hg.1 | ENST00000518530.2 | Ensembl |
| Affymetrix Human Clariom D Assay | TC0800006475.hg.1 | ENST00000518530.2 | Ensembl |
| Affymetrix Human Clariom D Assay | TC0800006475.hg.1 | ENST00000518530.2 | Ensembl |
| Affymetrix Human Clariom D Assay | TC0800006475.hg.1 | ENST00000518530.2 | Ensembl |
| Affymetrix Human Clariom D Assay | TC0800006475.hg.1 | ENST00000520901.5 | Ensembl |
| Affymetrix Human Clariom D Assay | TC0800006475.hg.1 | ENST00000520901.5 | Ensembl |
| Affymetrix Human Clariom D Assay | TC0800006475.hg.1 | ENST00000520901.5 | Ensembl |
| Affymetrix Human Clariom D Assay | TC0800006475.hg.1 | ENST00000520901.5 | Ensembl |
| Affymetrix Human Clariom D Assay | TC0800006475.hg.1 | ENST00000520901.5 | Ensembl |
| Affymetrix Human Clariom D Assay | TC0800006475.hg.1 | ENST00000520901.5 | Ensembl |
| Affymetrix Human Clariom D Assay | TC0800006475.hg.1 | ENST00000520901.5 | Ensembl |
| Affymetrix Human Clariom D Assay | TC0800006475.hg.1 | ENST00000520901.5 | Ensembl |
| Affymetrix Human Clariom D Assay | TC0800006475.hg.1 | ENST00000520901.5 | Ensembl |
| Affymetrix Human Clariom D Assay | TC0800006475.hg.1 | ENST00000520901.5 | Ensembl |
| Affymetrix Human Clariom D Assay | TC0800006475.hg.1 | ENST00000520901.5 | Ensembl |
| Affymetrix Human Clariom D Assay | TC0800006475.hg.1 | ENST00000520901.5 | Ensembl |
| Affymetrix Human Clariom D Assay | TC0800006475.hg.1 | ENST00000520901.5 | Ensembl |
| Affymetrix Human Clariom D Assay | TC0800006475.hg.1 | ENST00000520901.5 | Ensembl |
| Affymetrix Human Clariom D Assay | TC0800006475.hg.1 | ENST00000522499.1 | Ensembl |
| Affymetrix Human Clariom D Assay | TC0800006475.hg.1 | ENST00000522499.1 | Ensembl |
| Affymetrix Human Clariom D Assay | TC0800006475.hg.1 | ENST00000522499.1 | Ensembl |
| Affymetrix Human Clariom D Assay | TC0800006475.hg.1 | ENST00000522499.1 | Ensembl |
| Affymetrix Human Clariom D Assay | TC0800006475.hg.1 | ENST00000522499.1 | Ensembl |
| Affymetrix Human Clariom D Assay | TC0800006475.hg.1 | ENST00000524065.4 | Ensembl |
| Affymetrix Human Clariom D Assay | TC0800006475.hg.1 | ENST00000524065.4 | Ensembl |
| Affymetrix Human Clariom D Assay | TC0800006475.hg.1 | ENST00000524065.4 | Ensembl |
| Affymetrix Human Clariom D Assay | TC0800006475.hg.1 | ENST00000524065.4 | Ensembl |
| Affymetrix Human Clariom D Assay | TC0800006475.hg.1 | ENST00000524065.4 | Ensembl |
| Affymetrix Human Clariom D Assay | TC0800006475.hg.1 | ENST00000612087.1 | Ensembl |
| Affymetrix Human Clariom D Assay | TC0800006475.hg.1 | ENST00000612087.1 | Ensembl |
| Affymetrix Human Clariom D Assay | TC0800006475.hg.1 | ENST00000612087.1 | Ensembl |
| Affymetrix Human Clariom D Assay | TC0800006475.hg.1 | ENST00000612087.1 | Ensembl |
| Affymetrix Human Clariom D Assay | TC0800006475.hg.1 | ENST00000612087.1 | Ensembl |
| Affymetrix Human Clariom D Assay | TC0800006475.hg.1 | ENST00000612087.1 | Ensembl |
| Affymetrix Human Clariom D Assay | TC0800006475.hg.1 | ENST00000612087.1 | Ensembl |
| Affymetrix Human Clariom D Assay | TC0800006475.hg.1 | ENST00000612087.1 | Ensembl |
| Affymetrix Human Clariom D Assay | TC0800006475.hg.1 | ENST00000612087.1 | Ensembl |
| Affymetrix Human Clariom D Assay | TC0800006475.hg.1 | ENST00000612087.1 | Ensembl |
| Affymetrix Human Clariom D Assay | TC0800006475.hg.1 | ENST00000612087.1 | Ensembl |
| Affymetrix Human Clariom D Assay | TC0800006475.hg.1 | ENST00000612087.1 | Ensembl |
| Affymetrix Human Clariom D Assay | TC0800006475.hg.1 | ENST00000612087.1 | Ensembl |
| Affymetrix Human Clariom D Assay | TC0800006475.hg.1 | ENST00000612087.1 | Ensembl |
| Affymetrix Human Clariom D Assay | TC0800006475.hg.1 | ENST00000612087.1 | Ensembl |
| Affymetrix Human Clariom D Assay | TC0800006475.hg.1 | ENST00000612087.1 | Ensembl |
| Affymetrix Human Clariom D Assay | TC0800006475.hg.1 | ENST00000612087.1 | Ensembl |
| Affymetrix Human Clariom D Assay | TC0800006475.hg.1 | NM_001277161.1 | Refseq |
| Affymetrix Human Clariom D Assay | TC0800006475.hg.1 | NM_001277161.1 | Refseq |
| Affymetrix Human Clariom D Assay | TC0800006475.hg.1 | NM_001277161.1 | Refseq |
| Affymetrix Human Clariom D Assay | TC0800006475.hg.1 | NM_001277161.1 | Refseq |
| Affymetrix Human Clariom D Assay | TC0800006475.hg.1 | NM_001277161.1 | Refseq |
| Affymetrix Human Clariom D Assay | TC0800006475.hg.1 | NM_001277161.1 | Refseq |
| Affymetrix Human Clariom D Assay | TC0800006475.hg.1 | NM_001277161.1 | Refseq |
| Affymetrix Human Clariom D Assay | TC0800006475.hg.1 | NM_001277161.1 | Refseq |
| Affymetrix Human Clariom D Assay | TC0800006475.hg.1 | NM_001277161.1 | Refseq |
| Affymetrix Human Clariom D Assay | TC0800006475.hg.1 | NM_001277161.1 | Refseq |
| Affymetrix Human Clariom D Assay | TC0800006475.hg.1 | NM_001277161.1 | Refseq |
| Affymetrix Human Clariom D Assay | TC0800006475.hg.1 | NM_001277161.1 | Refseq |
| Affymetrix Human Clariom D Assay | TC0800006475.hg.1 | NM_001277161.1 | Refseq |
| Affymetrix Human Clariom D Assay | TC0800006475.hg.1 | NM_001277161.1 | Refseq |
| Affymetrix Human Clariom D Assay | TC0800006475.hg.1 | NM_001277161.1 | Refseq |
| Affymetrix Human Clariom D Assay | TC0800006475.hg.1 | NM_001277161.1 | Refseq |
| Affymetrix Human Clariom D Assay | TC0800006475.hg.1 | NM_001277161.1 | Refseq |
| Affymetrix Human Clariom D Assay | TC0800006475.hg.1 | NM_004745.4 | Refseq |
| Affymetrix Human Clariom D Assay | TC0800006475.hg.1 | NM_004745.4 | Refseq |
| Affymetrix Human Clariom D Assay | TC0800006475.hg.1 | NM_004745.4 | Refseq |
| Affymetrix Human Clariom D Assay | TC0800006475.hg.1 | NM_004745.4 | Refseq |
| Affymetrix Human Clariom D Assay | TC0800006475.hg.1 | NM_004745.4 | Refseq |
| Affymetrix Human Clariom D Assay | TC0800006475.hg.1 | NM_004745.4 | Refseq |
| Affymetrix Human Clariom D Assay | TC0800006475.hg.1 | NM_004745.4 | Refseq |
| Affymetrix Human Clariom D Assay | TC0800006475.hg.1 | NM_004745.4 | Refseq |
| Affymetrix Human Clariom D Assay | TC0800006475.hg.1 | NM_004745.4 | Refseq |
| Affymetrix Human Clariom D Assay | TC0800006475.hg.1 | NM_004745.4 | Refseq |
| Affymetrix Human Clariom D Assay | TC0800006475.hg.1 | NM_004745.4 | Refseq |
| Affymetrix Human Clariom D Assay | TC0800006475.hg.1 | NM_004745.4 | Refseq |
| Affymetrix Human Clariom D Assay | TC0800006475.hg.1 | NM_004745.4 | Refseq |
| Affymetrix Human Clariom D Assay | TC0800006475.hg.1 | NM_004745.4 | Refseq |
| Affymetrix Human Clariom D Assay | TC0800006475.hg.1 | NM_004745.4 | Refseq |
| Affymetrix Human Clariom D Assay | TC0800006475.hg.1 | NM_004745.4 | Refseq |
| Affymetrix Human Clariom D Assay | TC0800006475.hg.1 | NM_004745.4 | Refseq |
| Affymetrix Human Clariom D Assay | TC0800006475.hg.1 | NM_004745.4 | Refseq |
| Affymetrix Human Clariom D Assay | PSR0800144291.hg.1 | ENST00000421627.6 | Ensembl |
| Affymetrix Human Clariom D Assay | PSR0800144291.hg.1 | NM_004745.4 | Refseq |
| Affymetrix Human Clariom D Assay | PSR0800144291.hg.1 | ENST00000421627.6 | Ensembl |
| Affymetrix Human Clariom D Assay | PSR0800144291.hg.1 | NM_004745.4 | Refseq |
| Affymetrix Human Clariom D Assay | PSR0800144291.hg.1 | TC0800006475.hg.1 | Clariom D TC |

DLGAP2
Genomic Map