Assay Details
| Gene Symbol: DLGAP2 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | NM_004745.4 | NP_004736.2 | 7 - 8 | 2092 | 64 | |
| GenBank mRNA | AB000275.1 | - | 4 - 5 | 647 | 64 | |
| AF119817.1 | - | 7 - 8 | 2077 | 64 | ||
| AF119818.1 | - | 7 - 8 | 2092 | 64 | ||
| BC141862.1 | - | 6 - 7 | 2008 | 40027523 | 64 | |
| KJ534823.1 | - | 6 - 7 | 1958 | 64 | ||
More Information
Updated Mapping Information
Set Membership: |
|
Matching Ambion siRNAs:
Panther Classification:
Gene Ontology Categories:
MicroArray Probes:
Affymetrix
| Array Name | Probe IDs | Targets | Target Type |
|---|---|---|---|
| Affymetrix Human Transcriptome Array 2.0 | PSR08000109.hg.1 | TC08000007.hg.1 | HTA TC |
| Affymetrix Human Transcriptome Array 2.0 | PSR08000109.hg.1 | ENST00000356067.4 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | PSR08000109.hg.1 | ENST00000421627.2 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | PSR08000109.hg.1 | NM_004745.4 | Refseq |
| Affymetrix Human Transcriptome Array 2.0 | TC0800006475.hg.1 | ENST00000421627.6 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0800006475.hg.1 | ENST00000421627.6 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0800006475.hg.1 | ENST00000421627.6 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0800006475.hg.1 | ENST00000421627.6 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0800006475.hg.1 | ENST00000421627.6 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0800006475.hg.1 | ENST00000421627.6 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0800006475.hg.1 | ENST00000421627.6 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0800006475.hg.1 | ENST00000421627.6 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0800006475.hg.1 | ENST00000421627.6 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0800006475.hg.1 | ENST00000421627.6 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0800006475.hg.1 | ENST00000421627.6 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0800006475.hg.1 | ENST00000421627.6 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0800006475.hg.1 | ENST00000421627.6 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0800006475.hg.1 | ENST00000421627.6 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0800006475.hg.1 | ENST00000421627.6 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0800006475.hg.1 | ENST00000421627.6 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0800006475.hg.1 | ENST00000421627.6 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0800006475.hg.1 | ENST00000421627.6 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0800006475.hg.1 | ENST00000518530.2 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0800006475.hg.1 | ENST00000518530.2 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0800006475.hg.1 | ENST00000518530.2 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0800006475.hg.1 | ENST00000518530.2 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0800006475.hg.1 | ENST00000520901.5 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0800006475.hg.1 | ENST00000520901.5 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0800006475.hg.1 | ENST00000520901.5 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0800006475.hg.1 | ENST00000520901.5 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0800006475.hg.1 | ENST00000520901.5 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0800006475.hg.1 | ENST00000520901.5 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0800006475.hg.1 | ENST00000520901.5 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0800006475.hg.1 | ENST00000520901.5 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0800006475.hg.1 | ENST00000520901.5 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0800006475.hg.1 | ENST00000520901.5 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0800006475.hg.1 | ENST00000520901.5 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0800006475.hg.1 | ENST00000520901.5 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0800006475.hg.1 | ENST00000520901.5 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0800006475.hg.1 | ENST00000520901.5 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0800006475.hg.1 | ENST00000522499.1 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0800006475.hg.1 | ENST00000522499.1 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0800006475.hg.1 | ENST00000522499.1 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0800006475.hg.1 | ENST00000522499.1 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0800006475.hg.1 | ENST00000522499.1 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0800006475.hg.1 | ENST00000524065.4 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0800006475.hg.1 | ENST00000524065.4 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0800006475.hg.1 | ENST00000524065.4 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0800006475.hg.1 | ENST00000524065.4 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0800006475.hg.1 | ENST00000524065.4 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0800006475.hg.1 | ENST00000612087.1 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0800006475.hg.1 | ENST00000612087.1 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0800006475.hg.1 | ENST00000612087.1 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0800006475.hg.1 | ENST00000612087.1 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0800006475.hg.1 | ENST00000612087.1 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0800006475.hg.1 | ENST00000612087.1 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0800006475.hg.1 | ENST00000612087.1 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0800006475.hg.1 | ENST00000612087.1 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0800006475.hg.1 | ENST00000612087.1 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0800006475.hg.1 | ENST00000612087.1 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0800006475.hg.1 | ENST00000612087.1 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0800006475.hg.1 | ENST00000612087.1 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0800006475.hg.1 | ENST00000612087.1 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0800006475.hg.1 | ENST00000612087.1 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0800006475.hg.1 | ENST00000612087.1 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0800006475.hg.1 | ENST00000612087.1 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0800006475.hg.1 | ENST00000612087.1 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0800006475.hg.1 | NM_001277161.1 | Refseq |
| Affymetrix Human Transcriptome Array 2.0 | TC0800006475.hg.1 | NM_001277161.1 | Refseq |
| Affymetrix Human Transcriptome Array 2.0 | TC0800006475.hg.1 | NM_001277161.1 | Refseq |
| Affymetrix Human Transcriptome Array 2.0 | TC0800006475.hg.1 | NM_001277161.1 | Refseq |
| Affymetrix Human Transcriptome Array 2.0 | TC0800006475.hg.1 | NM_001277161.1 | Refseq |
| Affymetrix Human Transcriptome Array 2.0 | TC0800006475.hg.1 | NM_001277161.1 | Refseq |
| Affymetrix Human Transcriptome Array 2.0 | TC0800006475.hg.1 | NM_001277161.1 | Refseq |
| Affymetrix Human Transcriptome Array 2.0 | TC0800006475.hg.1 | NM_001277161.1 | Refseq |
| Affymetrix Human Transcriptome Array 2.0 | TC0800006475.hg.1 | NM_001277161.1 | Refseq |
| Affymetrix Human Transcriptome Array 2.0 | TC0800006475.hg.1 | NM_001277161.1 | Refseq |
| Affymetrix Human Transcriptome Array 2.0 | TC0800006475.hg.1 | NM_001277161.1 | Refseq |
| Affymetrix Human Transcriptome Array 2.0 | TC0800006475.hg.1 | NM_001277161.1 | Refseq |
| Affymetrix Human Transcriptome Array 2.0 | TC0800006475.hg.1 | NM_001277161.1 | Refseq |
| Affymetrix Human Transcriptome Array 2.0 | TC0800006475.hg.1 | NM_001277161.1 | Refseq |
| Affymetrix Human Transcriptome Array 2.0 | TC0800006475.hg.1 | NM_001277161.1 | Refseq |
| Affymetrix Human Transcriptome Array 2.0 | TC0800006475.hg.1 | NM_001277161.1 | Refseq |
| Affymetrix Human Transcriptome Array 2.0 | TC0800006475.hg.1 | NM_001277161.1 | Refseq |
| Affymetrix Human Transcriptome Array 2.0 | TC0800006475.hg.1 | NM_004745.4 | Refseq |
| Affymetrix Human Transcriptome Array 2.0 | TC0800006475.hg.1 | NM_004745.4 | Refseq |
| Affymetrix Human Transcriptome Array 2.0 | TC0800006475.hg.1 | NM_004745.4 | Refseq |
| Affymetrix Human Transcriptome Array 2.0 | TC0800006475.hg.1 | NM_004745.4 | Refseq |
| Affymetrix Human Transcriptome Array 2.0 | TC0800006475.hg.1 | NM_004745.4 | Refseq |
| Affymetrix Human Transcriptome Array 2.0 | TC0800006475.hg.1 | NM_004745.4 | Refseq |
| Affymetrix Human Transcriptome Array 2.0 | TC0800006475.hg.1 | NM_004745.4 | Refseq |
| Affymetrix Human Transcriptome Array 2.0 | TC0800006475.hg.1 | NM_004745.4 | Refseq |
| Affymetrix Human Transcriptome Array 2.0 | TC0800006475.hg.1 | NM_004745.4 | Refseq |
| Affymetrix Human Transcriptome Array 2.0 | TC0800006475.hg.1 | NM_004745.4 | Refseq |
| Affymetrix Human Transcriptome Array 2.0 | TC0800006475.hg.1 | NM_004745.4 | Refseq |
| Affymetrix Human Transcriptome Array 2.0 | TC0800006475.hg.1 | NM_004745.4 | Refseq |
| Affymetrix Human Transcriptome Array 2.0 | TC0800006475.hg.1 | NM_004745.4 | Refseq |
| Affymetrix Human Transcriptome Array 2.0 | TC0800006475.hg.1 | NM_004745.4 | Refseq |
| Affymetrix Human Transcriptome Array 2.0 | TC0800006475.hg.1 | NM_004745.4 | Refseq |
| Affymetrix Human Transcriptome Array 2.0 | TC0800006475.hg.1 | NM_004745.4 | Refseq |
| Affymetrix Human Transcriptome Array 2.0 | TC0800006475.hg.1 | NM_004745.4 | Refseq |
| Affymetrix Human Transcriptome Array 2.0 | TC0800006475.hg.1 | NM_004745.4 | Refseq |
| Affymetrix Human Clariom D Assay | TC0800006475.hg.1 | ENST00000421627.6 | Ensembl |
| Affymetrix Human Clariom D Assay | TC0800006475.hg.1 | ENST00000421627.6 | Ensembl |
| Affymetrix Human Clariom D Assay | TC0800006475.hg.1 | ENST00000421627.6 | Ensembl |
| Affymetrix Human Clariom D Assay | TC0800006475.hg.1 | ENST00000421627.6 | Ensembl |
| Affymetrix Human Clariom D Assay | TC0800006475.hg.1 | ENST00000421627.6 | Ensembl |
| Affymetrix Human Clariom D Assay | TC0800006475.hg.1 | ENST00000421627.6 | Ensembl |
| Affymetrix Human Clariom D Assay | TC0800006475.hg.1 | ENST00000421627.6 | Ensembl |
| Affymetrix Human Clariom D Assay | TC0800006475.hg.1 | ENST00000421627.6 | Ensembl |
| Affymetrix Human Clariom D Assay | TC0800006475.hg.1 | ENST00000421627.6 | Ensembl |
| Affymetrix Human Clariom D Assay | TC0800006475.hg.1 | ENST00000421627.6 | Ensembl |
| Affymetrix Human Clariom D Assay | TC0800006475.hg.1 | ENST00000421627.6 | Ensembl |
| Affymetrix Human Clariom D Assay | TC0800006475.hg.1 | ENST00000421627.6 | Ensembl |
| Affymetrix Human Clariom D Assay | TC0800006475.hg.1 | ENST00000421627.6 | Ensembl |
| Affymetrix Human Clariom D Assay | TC0800006475.hg.1 | ENST00000421627.6 | Ensembl |
| Affymetrix Human Clariom D Assay | TC0800006475.hg.1 | ENST00000421627.6 | Ensembl |
| Affymetrix Human Clariom D Assay | TC0800006475.hg.1 | ENST00000421627.6 | Ensembl |
| Affymetrix Human Clariom D Assay | TC0800006475.hg.1 | ENST00000421627.6 | Ensembl |
| Affymetrix Human Clariom D Assay | TC0800006475.hg.1 | ENST00000421627.6 | Ensembl |
| Affymetrix Human Clariom D Assay | TC0800006475.hg.1 | ENST00000518530.2 | Ensembl |
| Affymetrix Human Clariom D Assay | TC0800006475.hg.1 | ENST00000518530.2 | Ensembl |
| Affymetrix Human Clariom D Assay | TC0800006475.hg.1 | ENST00000518530.2 | Ensembl |
| Affymetrix Human Clariom D Assay | TC0800006475.hg.1 | ENST00000518530.2 | Ensembl |
| Affymetrix Human Clariom D Assay | TC0800006475.hg.1 | ENST00000520901.5 | Ensembl |
| Affymetrix Human Clariom D Assay | TC0800006475.hg.1 | ENST00000520901.5 | Ensembl |
| Affymetrix Human Clariom D Assay | TC0800006475.hg.1 | ENST00000520901.5 | Ensembl |
| Affymetrix Human Clariom D Assay | TC0800006475.hg.1 | ENST00000520901.5 | Ensembl |
| Affymetrix Human Clariom D Assay | TC0800006475.hg.1 | ENST00000520901.5 | Ensembl |
| Affymetrix Human Clariom D Assay | TC0800006475.hg.1 | ENST00000520901.5 | Ensembl |
| Affymetrix Human Clariom D Assay | TC0800006475.hg.1 | ENST00000520901.5 | Ensembl |
| Affymetrix Human Clariom D Assay | TC0800006475.hg.1 | ENST00000520901.5 | Ensembl |
| Affymetrix Human Clariom D Assay | TC0800006475.hg.1 | ENST00000520901.5 | Ensembl |
| Affymetrix Human Clariom D Assay | TC0800006475.hg.1 | ENST00000520901.5 | Ensembl |
| Affymetrix Human Clariom D Assay | TC0800006475.hg.1 | ENST00000520901.5 | Ensembl |
| Affymetrix Human Clariom D Assay | TC0800006475.hg.1 | ENST00000520901.5 | Ensembl |
| Affymetrix Human Clariom D Assay | TC0800006475.hg.1 | ENST00000520901.5 | Ensembl |
| Affymetrix Human Clariom D Assay | TC0800006475.hg.1 | ENST00000520901.5 | Ensembl |
| Affymetrix Human Clariom D Assay | TC0800006475.hg.1 | ENST00000522499.1 | Ensembl |
| Affymetrix Human Clariom D Assay | TC0800006475.hg.1 | ENST00000522499.1 | Ensembl |
| Affymetrix Human Clariom D Assay | TC0800006475.hg.1 | ENST00000522499.1 | Ensembl |
| Affymetrix Human Clariom D Assay | TC0800006475.hg.1 | ENST00000522499.1 | Ensembl |
| Affymetrix Human Clariom D Assay | TC0800006475.hg.1 | ENST00000522499.1 | Ensembl |
| Affymetrix Human Clariom D Assay | TC0800006475.hg.1 | ENST00000524065.4 | Ensembl |
| Affymetrix Human Clariom D Assay | TC0800006475.hg.1 | ENST00000524065.4 | Ensembl |
| Affymetrix Human Clariom D Assay | TC0800006475.hg.1 | ENST00000524065.4 | Ensembl |
| Affymetrix Human Clariom D Assay | TC0800006475.hg.1 | ENST00000524065.4 | Ensembl |
| Affymetrix Human Clariom D Assay | TC0800006475.hg.1 | ENST00000524065.4 | Ensembl |
| Affymetrix Human Clariom D Assay | TC0800006475.hg.1 | ENST00000612087.1 | Ensembl |
| Affymetrix Human Clariom D Assay | TC0800006475.hg.1 | ENST00000612087.1 | Ensembl |
| Affymetrix Human Clariom D Assay | TC0800006475.hg.1 | ENST00000612087.1 | Ensembl |
| Affymetrix Human Clariom D Assay | TC0800006475.hg.1 | ENST00000612087.1 | Ensembl |
| Affymetrix Human Clariom D Assay | TC0800006475.hg.1 | ENST00000612087.1 | Ensembl |
| Affymetrix Human Clariom D Assay | TC0800006475.hg.1 | ENST00000612087.1 | Ensembl |
| Affymetrix Human Clariom D Assay | TC0800006475.hg.1 | ENST00000612087.1 | Ensembl |
| Affymetrix Human Clariom D Assay | TC0800006475.hg.1 | ENST00000612087.1 | Ensembl |
| Affymetrix Human Clariom D Assay | TC0800006475.hg.1 | ENST00000612087.1 | Ensembl |
| Affymetrix Human Clariom D Assay | TC0800006475.hg.1 | ENST00000612087.1 | Ensembl |
| Affymetrix Human Clariom D Assay | TC0800006475.hg.1 | ENST00000612087.1 | Ensembl |
| Affymetrix Human Clariom D Assay | TC0800006475.hg.1 | ENST00000612087.1 | Ensembl |
| Affymetrix Human Clariom D Assay | TC0800006475.hg.1 | ENST00000612087.1 | Ensembl |
| Affymetrix Human Clariom D Assay | TC0800006475.hg.1 | ENST00000612087.1 | Ensembl |
| Affymetrix Human Clariom D Assay | TC0800006475.hg.1 | ENST00000612087.1 | Ensembl |
| Affymetrix Human Clariom D Assay | TC0800006475.hg.1 | ENST00000612087.1 | Ensembl |
| Affymetrix Human Clariom D Assay | TC0800006475.hg.1 | ENST00000612087.1 | Ensembl |
| Affymetrix Human Clariom D Assay | TC0800006475.hg.1 | NM_001277161.1 | Refseq |
| Affymetrix Human Clariom D Assay | TC0800006475.hg.1 | NM_001277161.1 | Refseq |
| Affymetrix Human Clariom D Assay | TC0800006475.hg.1 | NM_001277161.1 | Refseq |
| Affymetrix Human Clariom D Assay | TC0800006475.hg.1 | NM_001277161.1 | Refseq |
| Affymetrix Human Clariom D Assay | TC0800006475.hg.1 | NM_001277161.1 | Refseq |
| Affymetrix Human Clariom D Assay | TC0800006475.hg.1 | NM_001277161.1 | Refseq |
| Affymetrix Human Clariom D Assay | TC0800006475.hg.1 | NM_001277161.1 | Refseq |
| Affymetrix Human Clariom D Assay | TC0800006475.hg.1 | NM_001277161.1 | Refseq |
| Affymetrix Human Clariom D Assay | TC0800006475.hg.1 | NM_001277161.1 | Refseq |
| Affymetrix Human Clariom D Assay | TC0800006475.hg.1 | NM_001277161.1 | Refseq |
| Affymetrix Human Clariom D Assay | TC0800006475.hg.1 | NM_001277161.1 | Refseq |
| Affymetrix Human Clariom D Assay | TC0800006475.hg.1 | NM_001277161.1 | Refseq |
| Affymetrix Human Clariom D Assay | TC0800006475.hg.1 | NM_001277161.1 | Refseq |
| Affymetrix Human Clariom D Assay | TC0800006475.hg.1 | NM_001277161.1 | Refseq |
| Affymetrix Human Clariom D Assay | TC0800006475.hg.1 | NM_001277161.1 | Refseq |
| Affymetrix Human Clariom D Assay | TC0800006475.hg.1 | NM_001277161.1 | Refseq |
| Affymetrix Human Clariom D Assay | TC0800006475.hg.1 | NM_001277161.1 | Refseq |
| Affymetrix Human Clariom D Assay | TC0800006475.hg.1 | NM_004745.4 | Refseq |
| Affymetrix Human Clariom D Assay | TC0800006475.hg.1 | NM_004745.4 | Refseq |
| Affymetrix Human Clariom D Assay | TC0800006475.hg.1 | NM_004745.4 | Refseq |
| Affymetrix Human Clariom D Assay | TC0800006475.hg.1 | NM_004745.4 | Refseq |
| Affymetrix Human Clariom D Assay | TC0800006475.hg.1 | NM_004745.4 | Refseq |
| Affymetrix Human Clariom D Assay | TC0800006475.hg.1 | NM_004745.4 | Refseq |
| Affymetrix Human Clariom D Assay | TC0800006475.hg.1 | NM_004745.4 | Refseq |
| Affymetrix Human Clariom D Assay | TC0800006475.hg.1 | NM_004745.4 | Refseq |
| Affymetrix Human Clariom D Assay | TC0800006475.hg.1 | NM_004745.4 | Refseq |
| Affymetrix Human Clariom D Assay | TC0800006475.hg.1 | NM_004745.4 | Refseq |
| Affymetrix Human Clariom D Assay | TC0800006475.hg.1 | NM_004745.4 | Refseq |
| Affymetrix Human Clariom D Assay | TC0800006475.hg.1 | NM_004745.4 | Refseq |
| Affymetrix Human Clariom D Assay | TC0800006475.hg.1 | NM_004745.4 | Refseq |
| Affymetrix Human Clariom D Assay | TC0800006475.hg.1 | NM_004745.4 | Refseq |
| Affymetrix Human Clariom D Assay | TC0800006475.hg.1 | NM_004745.4 | Refseq |
| Affymetrix Human Clariom D Assay | TC0800006475.hg.1 | NM_004745.4 | Refseq |
| Affymetrix Human Clariom D Assay | TC0800006475.hg.1 | NM_004745.4 | Refseq |
| Affymetrix Human Clariom D Assay | TC0800006475.hg.1 | NM_004745.4 | Refseq |

DLGAP2
Genomic Map