Assay Details
| Gene Symbol: FIG4 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | NM_014845.5 | NP_055660.1 | 14 - 15 | 1795 | 112 | |
| XM_011536281.2 | XP_011534583.1 | 14 - 15 | 2327 | 112 | ||
| XM_017011591.1 | XP_016867080.1 | 14 - 15 | 1800 | 112 | ||
| XM_017011592.1 | XP_016867081.1 | 13 - 14 | 1563 | 112 | ||
| XM_017011593.1 | XP_016867082.1 | 12 - 13 | 1498 | 112 | ||
| GenBank mRNA | AK222732.1 | - | 14 - 15 | 1692 | 112 | |
| AK291671.1 | - | 14 - 15 | 1720 | 112 | ||
| AK293422.1 | - | 9 - 10 | 1046 | 112 | ||
| BC041338.2 | - | 14 - 15 | 1751 | 5274004 | 112 | |
| D87464.1 | - | 14 - 15 | 1704 | 112 | ||
More Information
Updated Mapping Information
This assay targets the same location as HTA transcript cluster(s) TC06000879.hg.1
Set Membership: |
|
Matching Ambion siRNAs:
Panther Classification:
Gene Ontology Categories:
Function(s)
Process(es)
phosphatidylinositol biosynthetic process
vacuole organization
locomotory behavior
positive regulation of neuron projection development
dephosphorylation
negative regulation of myelination
myelin assembly
phosphatidylinositol-3-phosphate biosynthetic process
pigmentation
neuron development
phosphatidylinositol-3-phosphatase activity
protein binding
phosphatidylinositol-4-phosphate phosphatase activity
phosphatidylinositol-3,5-bisphosphate 5-phosphatase activity
phosphatidylinositol biosynthetic process
vacuole organization
locomotory behavior
positive regulation of neuron projection development
dephosphorylation
negative regulation of myelination
myelin assembly
phosphatidylinositol-3-phosphate biosynthetic process
pigmentation
neuron development
phosphatidylinositol-3-phosphatase activity
protein binding
phosphatidylinositol-4-phosphate phosphatase activity
phosphatidylinositol-3,5-bisphosphate 5-phosphatase activity
MicroArray Probes:
Affymetrix
| Array Name | Probe IDs | Targets | Target Type |
|---|---|---|---|
| Affymetrix Human Transcriptome Array 2.0 | TC0600009102.hg.1 | ENST00000230124.7 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0600009102.hg.1 | ENST00000230124.7 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0600009102.hg.1 | ENST00000230124.7 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0600009102.hg.1 | ENST00000230124.7 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0600009102.hg.1 | ENST00000230124.7 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0600009102.hg.1 | ENST00000230124.7 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0600009102.hg.1 | ENST00000230124.7 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0600009102.hg.1 | ENST00000230124.7 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0600009102.hg.1 | ENST00000230124.7 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0600009102.hg.1 | ENST00000230124.7 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0600009102.hg.1 | ENST00000230124.7 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0600009102.hg.1 | ENST00000230124.7 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0600009102.hg.1 | ENST00000230124.7 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0600009102.hg.1 | ENST00000230124.7 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0600009102.hg.1 | ENST00000230124.7 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0600009102.hg.1 | ENST00000230124.7 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0600009102.hg.1 | ENST00000230124.7 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0600009102.hg.1 | ENST00000230124.7 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0600009102.hg.1 | ENST00000230124.7 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0600009102.hg.1 | ENST00000230124.7 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0600009102.hg.1 | ENST00000230124.7 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0600009102.hg.1 | ENST00000230124.7 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0600009102.hg.1 | ENST00000230124.7 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0600009102.hg.1 | ENST00000230124.7 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0600009102.hg.1 | ENST00000230124.7 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0600009102.hg.1 | ENST00000230124.7 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0600009102.hg.1 | ENST00000368941.1 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0600009102.hg.1 | ENST00000368941.1 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0600009102.hg.1 | ENST00000368941.1 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0600009102.hg.1 | ENST00000368941.1 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0600009102.hg.1 | ENST00000368941.1 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0600009102.hg.1 | ENST00000368941.1 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0600009102.hg.1 | ENST00000415980.1 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0600009102.hg.1 | ENST00000415980.1 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0600009102.hg.1 | ENST00000415980.1 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0600009102.hg.1 | ENST00000415980.1 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0600009102.hg.1 | ENST00000419951.1 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0600009102.hg.1 | ENST00000419951.1 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0600009102.hg.1 | ENST00000419951.1 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0600009102.hg.1 | ENST00000419951.1 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0600009102.hg.1 | ENST00000454215.5 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0600009102.hg.1 | ENST00000454215.5 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0600009102.hg.1 | ENST00000454215.5 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0600009102.hg.1 | ENST00000454215.5 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0600009102.hg.1 | ENST00000454215.5 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0600009102.hg.1 | ENST00000454215.5 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0600009102.hg.1 | ENST00000454215.5 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0600009102.hg.1 | NM_014845.5 | Refseq |
| Affymetrix Human Transcriptome Array 2.0 | TC0600009102.hg.1 | NM_014845.5 | Refseq |
| Affymetrix Human Transcriptome Array 2.0 | TC0600009102.hg.1 | NM_014845.5 | Refseq |
| Affymetrix Human Transcriptome Array 2.0 | TC0600009102.hg.1 | NM_014845.5 | Refseq |
| Affymetrix Human Transcriptome Array 2.0 | TC0600009102.hg.1 | NM_014845.5 | Refseq |
| Affymetrix Human Transcriptome Array 2.0 | TC0600009102.hg.1 | NM_014845.5 | Refseq |
| Affymetrix Human Transcriptome Array 2.0 | TC0600009102.hg.1 | NM_014845.5 | Refseq |
| Affymetrix Human Transcriptome Array 2.0 | TC0600009102.hg.1 | NM_014845.5 | Refseq |
| Affymetrix Human Transcriptome Array 2.0 | TC0600009102.hg.1 | NM_014845.5 | Refseq |
| Affymetrix Human Transcriptome Array 2.0 | TC0600009102.hg.1 | NM_014845.5 | Refseq |
| Affymetrix Human Transcriptome Array 2.0 | TC0600009102.hg.1 | NM_014845.5 | Refseq |
| Affymetrix Human Transcriptome Array 2.0 | TC0600009102.hg.1 | NM_014845.5 | Refseq |
| Affymetrix Human Transcriptome Array 2.0 | TC0600009102.hg.1 | NM_014845.5 | Refseq |
| Affymetrix Human Transcriptome Array 2.0 | TC0600009102.hg.1 | NM_014845.5 | Refseq |
| Affymetrix Human Transcriptome Array 2.0 | TC0600009102.hg.1 | NM_014845.5 | Refseq |
| Affymetrix Human Transcriptome Array 2.0 | TC0600009102.hg.1 | NM_014845.5 | Refseq |
| Affymetrix Human Transcriptome Array 2.0 | TC0600009102.hg.1 | NM_014845.5 | Refseq |
| Affymetrix Human Transcriptome Array 2.0 | TC0600009102.hg.1 | NM_014845.5 | Refseq |
| Affymetrix Human Transcriptome Array 2.0 | TC0600009102.hg.1 | NM_014845.5 | Refseq |
| Affymetrix Human Transcriptome Array 2.0 | TC0600009102.hg.1 | NM_014845.5 | Refseq |
| Affymetrix Human Transcriptome Array 2.0 | TC0600009102.hg.1 | NM_014845.5 | Refseq |
| Affymetrix Human Transcriptome Array 2.0 | TC0600009102.hg.1 | NM_014845.5 | Refseq |
| Affymetrix Human Transcriptome Array 2.0 | TC0600009102.hg.1 | NM_014845.5 | Refseq |
| Affymetrix Human Transcriptome Array 2.0 | TC0600009102.hg.1 | NM_014845.5 | Refseq |
| Affymetrix Human Transcriptome Array 2.0 | TC0600009102.hg.1 | NM_014845.5 | Refseq |
| Affymetrix Human Transcriptome Array 2.0 | TC0600009102.hg.1 | NM_014845.5 | Refseq |
| Affymetrix Human Transcriptome Array 2.0 | TC0600009102.hg.1 | NM_014845.5 | Refseq |
| Affymetrix Human Transcriptome Array 2.0 | TC0600009102.hg.1 | NM_014845.5 | Refseq |
| Affymetrix Human Transcriptome Array 2.0 | PSR06010373.hg.1 | TC06000879.hg.1 | HTA TC |
| Affymetrix Human Transcriptome Array 2.0 | PSR06010373.hg.1 | ENST00000230124.3 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | PSR06010373.hg.1 | ENST00000441478.2 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | PSR06010373.hg.1 | NM_014845.5 | Refseq |
| Affymetrix Human Clariom D Assay | PSR0600162558.hg.1 | ENST00000230124.7 | Ensembl |
| Affymetrix Human Clariom D Assay | PSR0600162558.hg.1 | ENST00000415980.1 | Ensembl |
| Affymetrix Human Clariom D Assay | PSR0600162558.hg.1 | NM_014845.5 | Refseq |
| Affymetrix Human Clariom D Assay | PSR0600162558.hg.1 | ENST00000230124.7 | Ensembl |
| Affymetrix Human Clariom D Assay | PSR0600162558.hg.1 | ENST00000415980.1 | Ensembl |
| Affymetrix Human Clariom D Assay | PSR0600162558.hg.1 | NM_014845.5 | Refseq |
| Affymetrix Human Clariom D Assay | PSR0600162558.hg.1 | TC0600009102.hg.1 | Clariom D TC |
| Affymetrix Human Clariom D Assay | TC0600009102.hg.1 | ENST00000230124.7 | Ensembl |
| Affymetrix Human Clariom D Assay | TC0600009102.hg.1 | ENST00000230124.7 | Ensembl |
| Affymetrix Human Clariom D Assay | TC0600009102.hg.1 | ENST00000230124.7 | Ensembl |
| Affymetrix Human Clariom D Assay | TC0600009102.hg.1 | ENST00000230124.7 | Ensembl |
| Affymetrix Human Clariom D Assay | TC0600009102.hg.1 | ENST00000230124.7 | Ensembl |
| Affymetrix Human Clariom D Assay | TC0600009102.hg.1 | ENST00000230124.7 | Ensembl |
| Affymetrix Human Clariom D Assay | TC0600009102.hg.1 | ENST00000230124.7 | Ensembl |
| Affymetrix Human Clariom D Assay | TC0600009102.hg.1 | ENST00000230124.7 | Ensembl |
| Affymetrix Human Clariom D Assay | TC0600009102.hg.1 | ENST00000230124.7 | Ensembl |
| Affymetrix Human Clariom D Assay | TC0600009102.hg.1 | ENST00000230124.7 | Ensembl |
| Affymetrix Human Clariom D Assay | TC0600009102.hg.1 | ENST00000230124.7 | Ensembl |
| Affymetrix Human Clariom D Assay | TC0600009102.hg.1 | ENST00000230124.7 | Ensembl |
| Affymetrix Human Clariom D Assay | TC0600009102.hg.1 | ENST00000230124.7 | Ensembl |
| Affymetrix Human Clariom D Assay | TC0600009102.hg.1 | ENST00000230124.7 | Ensembl |
| Affymetrix Human Clariom D Assay | TC0600009102.hg.1 | ENST00000230124.7 | Ensembl |
| Affymetrix Human Clariom D Assay | TC0600009102.hg.1 | ENST00000230124.7 | Ensembl |
| Affymetrix Human Clariom D Assay | TC0600009102.hg.1 | ENST00000230124.7 | Ensembl |
| Affymetrix Human Clariom D Assay | TC0600009102.hg.1 | ENST00000230124.7 | Ensembl |
| Affymetrix Human Clariom D Assay | TC0600009102.hg.1 | ENST00000230124.7 | Ensembl |
| Affymetrix Human Clariom D Assay | TC0600009102.hg.1 | ENST00000230124.7 | Ensembl |
| Affymetrix Human Clariom D Assay | TC0600009102.hg.1 | ENST00000230124.7 | Ensembl |
| Affymetrix Human Clariom D Assay | TC0600009102.hg.1 | ENST00000230124.7 | Ensembl |
| Affymetrix Human Clariom D Assay | TC0600009102.hg.1 | ENST00000230124.7 | Ensembl |
| Affymetrix Human Clariom D Assay | TC0600009102.hg.1 | ENST00000230124.7 | Ensembl |
| Affymetrix Human Clariom D Assay | TC0600009102.hg.1 | ENST00000230124.7 | Ensembl |
| Affymetrix Human Clariom D Assay | TC0600009102.hg.1 | ENST00000230124.7 | Ensembl |
| Affymetrix Human Clariom D Assay | TC0600009102.hg.1 | ENST00000368941.1 | Ensembl |
| Affymetrix Human Clariom D Assay | TC0600009102.hg.1 | ENST00000368941.1 | Ensembl |
| Affymetrix Human Clariom D Assay | TC0600009102.hg.1 | ENST00000368941.1 | Ensembl |
| Affymetrix Human Clariom D Assay | TC0600009102.hg.1 | ENST00000368941.1 | Ensembl |
| Affymetrix Human Clariom D Assay | TC0600009102.hg.1 | ENST00000368941.1 | Ensembl |
| Affymetrix Human Clariom D Assay | TC0600009102.hg.1 | ENST00000368941.1 | Ensembl |
| Affymetrix Human Clariom D Assay | TC0600009102.hg.1 | ENST00000415980.1 | Ensembl |
| Affymetrix Human Clariom D Assay | TC0600009102.hg.1 | ENST00000415980.1 | Ensembl |
| Affymetrix Human Clariom D Assay | TC0600009102.hg.1 | ENST00000415980.1 | Ensembl |
| Affymetrix Human Clariom D Assay | TC0600009102.hg.1 | ENST00000415980.1 | Ensembl |
| Affymetrix Human Clariom D Assay | TC0600009102.hg.1 | ENST00000419951.1 | Ensembl |
| Affymetrix Human Clariom D Assay | TC0600009102.hg.1 | ENST00000419951.1 | Ensembl |
| Affymetrix Human Clariom D Assay | TC0600009102.hg.1 | ENST00000419951.1 | Ensembl |
| Affymetrix Human Clariom D Assay | TC0600009102.hg.1 | ENST00000419951.1 | Ensembl |
| Affymetrix Human Clariom D Assay | TC0600009102.hg.1 | ENST00000454215.5 | Ensembl |
| Affymetrix Human Clariom D Assay | TC0600009102.hg.1 | ENST00000454215.5 | Ensembl |
| Affymetrix Human Clariom D Assay | TC0600009102.hg.1 | ENST00000454215.5 | Ensembl |
| Affymetrix Human Clariom D Assay | TC0600009102.hg.1 | ENST00000454215.5 | Ensembl |
| Affymetrix Human Clariom D Assay | TC0600009102.hg.1 | ENST00000454215.5 | Ensembl |
| Affymetrix Human Clariom D Assay | TC0600009102.hg.1 | ENST00000454215.5 | Ensembl |
| Affymetrix Human Clariom D Assay | TC0600009102.hg.1 | ENST00000454215.5 | Ensembl |
| Affymetrix Human Clariom D Assay | TC0600009102.hg.1 | NM_014845.5 | Refseq |
| Affymetrix Human Clariom D Assay | TC0600009102.hg.1 | NM_014845.5 | Refseq |
| Affymetrix Human Clariom D Assay | TC0600009102.hg.1 | NM_014845.5 | Refseq |
| Affymetrix Human Clariom D Assay | TC0600009102.hg.1 | NM_014845.5 | Refseq |
| Affymetrix Human Clariom D Assay | TC0600009102.hg.1 | NM_014845.5 | Refseq |
| Affymetrix Human Clariom D Assay | TC0600009102.hg.1 | NM_014845.5 | Refseq |
| Affymetrix Human Clariom D Assay | TC0600009102.hg.1 | NM_014845.5 | Refseq |
| Affymetrix Human Clariom D Assay | TC0600009102.hg.1 | NM_014845.5 | Refseq |
| Affymetrix Human Clariom D Assay | TC0600009102.hg.1 | NM_014845.5 | Refseq |
| Affymetrix Human Clariom D Assay | TC0600009102.hg.1 | NM_014845.5 | Refseq |
| Affymetrix Human Clariom D Assay | TC0600009102.hg.1 | NM_014845.5 | Refseq |
| Affymetrix Human Clariom D Assay | TC0600009102.hg.1 | NM_014845.5 | Refseq |
| Affymetrix Human Clariom D Assay | TC0600009102.hg.1 | NM_014845.5 | Refseq |
| Affymetrix Human Clariom D Assay | TC0600009102.hg.1 | NM_014845.5 | Refseq |
| Affymetrix Human Clariom D Assay | TC0600009102.hg.1 | NM_014845.5 | Refseq |
| Affymetrix Human Clariom D Assay | TC0600009102.hg.1 | NM_014845.5 | Refseq |
| Affymetrix Human Clariom D Assay | TC0600009102.hg.1 | NM_014845.5 | Refseq |
| Affymetrix Human Clariom D Assay | TC0600009102.hg.1 | NM_014845.5 | Refseq |
| Affymetrix Human Clariom D Assay | TC0600009102.hg.1 | NM_014845.5 | Refseq |
| Affymetrix Human Clariom D Assay | TC0600009102.hg.1 | NM_014845.5 | Refseq |
| Affymetrix Human Clariom D Assay | TC0600009102.hg.1 | NM_014845.5 | Refseq |
| Affymetrix Human Clariom D Assay | TC0600009102.hg.1 | NM_014845.5 | Refseq |
| Affymetrix Human Clariom D Assay | TC0600009102.hg.1 | NM_014845.5 | Refseq |
| Affymetrix Human Clariom D Assay | TC0600009102.hg.1 | NM_014845.5 | Refseq |
| Affymetrix Human Clariom D Assay | TC0600009102.hg.1 | NM_014845.5 | Refseq |
| Affymetrix Human Clariom D Assay | TC0600009102.hg.1 | NM_014845.5 | Refseq |
| Affymetrix Human Clariom D Assay | TC0600009102.hg.1 | NM_014845.5 | Refseq |
| Affymetrix Human Clariom D Assay | TC0600009102.hg.1 | NM_014845.5 | Refseq |

FIG4
Genomic Map