| Array Name |
Probe IDs |
Targets |
Target Type |
| Affymetrix Human Transcriptome Array 2.0 |
PSR0X004478.hg.1 |
TC0X000341.hg.1 |
HTA TC |
| Affymetrix Human Transcriptome Array 2.0 |
PSR0X004478.hg.1 |
ENST00000374922.4 |
Ensembl |
| Affymetrix Human Transcriptome Array 2.0 |
PSR0X004478.hg.1 |
NR_015367.2 |
Refseq |
| Affymetrix Human Transcriptome Array 2.0 |
PSR0X004474.hg.1 |
TC0X000341.hg.1 |
HTA TC |
| Affymetrix Human Transcriptome Array 2.0 |
PSR0X004474.hg.1 |
ENST00000374922.4 |
Ensembl |
| Affymetrix Human Transcriptome Array 2.0 |
PSR0X004474.hg.1 |
NR_015367.2 |
Refseq |
| Affymetrix Human Transcriptome Array 2.0 |
PSR0X004476.hg.1 |
TC0X000341.hg.1 |
HTA TC |
| Affymetrix Human Transcriptome Array 2.0 |
PSR0X004476.hg.1 |
ENST00000374922.4 |
Ensembl |
| Affymetrix Human Transcriptome Array 2.0 |
PSR0X004476.hg.1 |
NR_015367.2 |
Refseq |
| Affymetrix Human Transcriptome Array 2.0 |
PSR0X004479.hg.1 |
TC0X000341.hg.1 |
HTA TC |
| Affymetrix Human Transcriptome Array 2.0 |
PSR0X004479.hg.1 |
ENST00000374922.4 |
Ensembl |
| Affymetrix Human Transcriptome Array 2.0 |
PSR0X004479.hg.1 |
NR_015367.2 |
Refseq |
| Affymetrix Human Transcriptome Array 2.0 |
TC0X00007397.hg.1 |
ENST00000374922.4 |
Ensembl |
| Affymetrix Human Transcriptome Array 2.0 |
TC0X00007397.hg.1 |
ENST00000374922.4 |
Ensembl |
| Affymetrix Human Transcriptome Array 2.0 |
TC0X00007397.hg.1 |
ENST00000374922.4 |
Ensembl |
| Affymetrix Human Transcriptome Array 2.0 |
TC0X00007397.hg.1 |
ENST00000374922.4 |
Ensembl |
| Affymetrix Human Transcriptome Array 2.0 |
TC0X00007397.hg.1 |
ENST00000374922.4 |
Ensembl |
| Affymetrix Human Transcriptome Array 2.0 |
TC0X00007397.hg.1 |
ENST00000374922.4 |
Ensembl |
| Affymetrix Human Transcriptome Array 2.0 |
TC0X00007397.hg.1 |
ENST00000374922.4 |
Ensembl |
| Affymetrix Human Transcriptome Array 2.0 |
TC0X00007397.hg.1 |
ENST00000374922.4 |
Ensembl |
| Affymetrix Human Transcriptome Array 2.0 |
TC0X00007397.hg.1 |
ENST00000374922.4 |
Ensembl |
| Affymetrix Human Transcriptome Array 2.0 |
TC0X00007397.hg.1 |
ENST00000374922.4 |
Ensembl |
| Affymetrix Human Transcriptome Array 2.0 |
TC0X00007397.hg.1 |
ENST00000374922.4 |
Ensembl |
| Affymetrix Human Transcriptome Array 2.0 |
TC0X00007397.hg.1 |
ENST00000374922.4 |
Ensembl |
| Affymetrix Human Transcriptome Array 2.0 |
TC0X00007397.hg.1 |
ENST00000374922.4 |
Ensembl |
| Affymetrix Human Transcriptome Array 2.0 |
TC0X00007397.hg.1 |
ENST00000374922.4 |
Ensembl |
| Affymetrix Human Transcriptome Array 2.0 |
TC0X00007397.hg.1 |
ENST00000374922.4 |
Ensembl |
| Affymetrix Human Transcriptome Array 2.0 |
TC0X00007397.hg.1 |
ENST00000374922.4 |
Ensembl |
| Affymetrix Human Transcriptome Array 2.0 |
TC0X00007397.hg.1 |
ENST00000374922.4 |
Ensembl |
| Affymetrix Human Transcriptome Array 2.0 |
TC0X00007397.hg.1 |
ENST00000374922.4 |
Ensembl |
| Affymetrix Human Transcriptome Array 2.0 |
TC0X00007397.hg.1 |
ENST00000374922.4 |
Ensembl |
| Affymetrix Human Transcriptome Array 2.0 |
TC0X00007397.hg.1 |
ENST00000374922.4 |
Ensembl |
| Affymetrix Human Transcriptome Array 2.0 |
TC0X00007397.hg.1 |
ENST00000374922.4 |
Ensembl |
| Affymetrix Human Transcriptome Array 2.0 |
TC0X00007397.hg.1 |
ENST00000374922.4 |
Ensembl |
| Affymetrix Human Transcriptome Array 2.0 |
TC0X00007397.hg.1 |
ENST00000423617.1 |
Ensembl |
| Affymetrix Human Transcriptome Array 2.0 |
TC0X00007397.hg.1 |
ENST00000423617.1 |
Ensembl |
| Affymetrix Human Transcriptome Array 2.0 |
TC0X00007397.hg.1 |
ENST00000423617.1 |
Ensembl |
| Affymetrix Human Transcriptome Array 2.0 |
TC0X00007397.hg.1 |
ENST00000423617.1 |
Ensembl |
| Affymetrix Human Transcriptome Array 2.0 |
TC0X00007397.hg.1 |
ENST00000423617.1 |
Ensembl |
| Affymetrix Human Transcriptome Array 2.0 |
TC0X00007397.hg.1 |
ENST00000423617.1 |
Ensembl |
| Affymetrix Human Transcriptome Array 2.0 |
TC0X00007397.hg.1 |
ENST00000451583.1 |
Ensembl |
| Affymetrix Human Transcriptome Array 2.0 |
TC0X00007397.hg.1 |
ENST00000451583.1 |
Ensembl |
| Affymetrix Human Transcriptome Array 2.0 |
TC0X00007397.hg.1 |
ENST00000451583.1 |
Ensembl |
| Affymetrix Human Transcriptome Array 2.0 |
TC0X00007397.hg.1 |
ENST00000451583.1 |
Ensembl |
| Affymetrix Human Transcriptome Array 2.0 |
TC0X00007397.hg.1 |
NR_015367.2 |
Refseq |
| Affymetrix Human Transcriptome Array 2.0 |
TC0X00007397.hg.1 |
NR_015367.2 |
Refseq |
| Affymetrix Human Transcriptome Array 2.0 |
TC0X00007397.hg.1 |
NR_015367.2 |
Refseq |
| Affymetrix Human Transcriptome Array 2.0 |
TC0X00007397.hg.1 |
NR_015367.2 |
Refseq |
| Affymetrix Human Transcriptome Array 2.0 |
TC0X00007397.hg.1 |
NR_015367.2 |
Refseq |
| Affymetrix Human Transcriptome Array 2.0 |
TC0X00007397.hg.1 |
NR_015367.2 |
Refseq |
| Affymetrix Human Transcriptome Array 2.0 |
TC0X00007397.hg.1 |
NR_015367.2 |
Refseq |
| Affymetrix Human Transcriptome Array 2.0 |
TC0X00007397.hg.1 |
NR_015367.2 |
Refseq |
| Affymetrix Human Transcriptome Array 2.0 |
PSR0X004477.hg.1 |
TC0X000341.hg.1 |
HTA TC |
| Affymetrix Human Transcriptome Array 2.0 |
PSR0X004477.hg.1 |
ENST00000374922.4 |
Ensembl |
| Affymetrix Human Transcriptome Array 2.0 |
PSR0X004477.hg.1 |
NR_015367.2 |
Refseq |
| Affymetrix Human Clariom D Assay |
PSR0X00150872.hg.1 |
ENST00000374922.4 |
Ensembl |
| Affymetrix Human Clariom D Assay |
PSR0X00150872.hg.1 |
ENST00000374922.4 |
Ensembl |
| Affymetrix Human Clariom D Assay |
PSR0X00150872.hg.1 |
ENST00000423617.1 |
Ensembl |
| Affymetrix Human Clariom D Assay |
PSR0X00150872.hg.1 |
ENST00000451583.1 |
Ensembl |
| Affymetrix Human Clariom D Assay |
PSR0X00150872.hg.1 |
NR_015367.2 |
Refseq |
| Affymetrix Human Clariom D Assay |
PSR0X00150872.hg.1 |
ENST00000374922.4 |
Ensembl |
| Affymetrix Human Clariom D Assay |
PSR0X00150872.hg.1 |
ENST00000374922.4 |
Ensembl |
| Affymetrix Human Clariom D Assay |
PSR0X00150872.hg.1 |
ENST00000423617.1 |
Ensembl |
| Affymetrix Human Clariom D Assay |
PSR0X00150872.hg.1 |
ENST00000451583.1 |
Ensembl |
| Affymetrix Human Clariom D Assay |
PSR0X00150872.hg.1 |
NR_015367.2 |
Refseq |
| Affymetrix Human Clariom D Assay |
PSR0X00150872.hg.1 |
TC0X00007397.hg.1 |
Clariom D TC |
| Affymetrix Human Clariom D Assay |
PSR0X00150870.hg.1 |
ENST00000374922.4 |
Ensembl |
| Affymetrix Human Clariom D Assay |
PSR0X00150870.hg.1 |
ENST00000374922.4 |
Ensembl |
| Affymetrix Human Clariom D Assay |
PSR0X00150870.hg.1 |
NR_015367.2 |
Refseq |
| Affymetrix Human Clariom D Assay |
PSR0X00150870.hg.1 |
ENST00000374922.4 |
Ensembl |
| Affymetrix Human Clariom D Assay |
PSR0X00150870.hg.1 |
ENST00000374922.4 |
Ensembl |
| Affymetrix Human Clariom D Assay |
PSR0X00150870.hg.1 |
NR_015367.2 |
Refseq |
| Affymetrix Human Clariom D Assay |
PSR0X00150870.hg.1 |
TC0X00007397.hg.1 |
Clariom D TC |
| Affymetrix Human Clariom D Assay |
PSR0X00150873.hg.1 |
ENST00000374922.4 |
Ensembl |
| Affymetrix Human Clariom D Assay |
PSR0X00150873.hg.1 |
ENST00000374922.4 |
Ensembl |
| Affymetrix Human Clariom D Assay |
PSR0X00150873.hg.1 |
ENST00000423617.1 |
Ensembl |
| Affymetrix Human Clariom D Assay |
PSR0X00150873.hg.1 |
NR_015367.2 |
Refseq |
| Affymetrix Human Clariom D Assay |
PSR0X00150873.hg.1 |
ENST00000374922.4 |
Ensembl |
| Affymetrix Human Clariom D Assay |
PSR0X00150873.hg.1 |
ENST00000374922.4 |
Ensembl |
| Affymetrix Human Clariom D Assay |
PSR0X00150873.hg.1 |
ENST00000423617.1 |
Ensembl |
| Affymetrix Human Clariom D Assay |
PSR0X00150873.hg.1 |
NR_015367.2 |
Refseq |
| Affymetrix Human Clariom D Assay |
PSR0X00150873.hg.1 |
TC0X00007397.hg.1 |
Clariom D TC |
| Affymetrix Human Clariom D Assay |
PSR0X00150875.hg.1 |
ENST00000374922.4 |
Ensembl |
| Affymetrix Human Clariom D Assay |
PSR0X00150875.hg.1 |
ENST00000374922.4 |
Ensembl |
| Affymetrix Human Clariom D Assay |
PSR0X00150875.hg.1 |
ENST00000451583.1 |
Ensembl |
| Affymetrix Human Clariom D Assay |
PSR0X00150875.hg.1 |
NR_015367.2 |
Refseq |
| Affymetrix Human Clariom D Assay |
PSR0X00150875.hg.1 |
ENST00000374922.4 |
Ensembl |
| Affymetrix Human Clariom D Assay |
PSR0X00150875.hg.1 |
ENST00000374922.4 |
Ensembl |
| Affymetrix Human Clariom D Assay |
PSR0X00150875.hg.1 |
ENST00000451583.1 |
Ensembl |
| Affymetrix Human Clariom D Assay |
PSR0X00150875.hg.1 |
NR_015367.2 |
Refseq |
| Affymetrix Human Clariom D Assay |
PSR0X00150875.hg.1 |
TC0X00007397.hg.1 |
Clariom D TC |
| Affymetrix Human Clariom D Assay |
TC0X00007397.hg.1 |
ENST00000374922.4 |
Ensembl |
| Affymetrix Human Clariom D Assay |
TC0X00007397.hg.1 |
ENST00000374922.4 |
Ensembl |
| Affymetrix Human Clariom D Assay |
TC0X00007397.hg.1 |
ENST00000374922.4 |
Ensembl |
| Affymetrix Human Clariom D Assay |
TC0X00007397.hg.1 |
ENST00000374922.4 |
Ensembl |
| Affymetrix Human Clariom D Assay |
TC0X00007397.hg.1 |
ENST00000374922.4 |
Ensembl |
| Affymetrix Human Clariom D Assay |
TC0X00007397.hg.1 |
ENST00000374922.4 |
Ensembl |
| Affymetrix Human Clariom D Assay |
TC0X00007397.hg.1 |
ENST00000374922.4 |
Ensembl |
| Affymetrix Human Clariom D Assay |
TC0X00007397.hg.1 |
ENST00000374922.4 |
Ensembl |
| Affymetrix Human Clariom D Assay |
TC0X00007397.hg.1 |
ENST00000374922.4 |
Ensembl |
| Affymetrix Human Clariom D Assay |
TC0X00007397.hg.1 |
ENST00000374922.4 |
Ensembl |
| Affymetrix Human Clariom D Assay |
TC0X00007397.hg.1 |
ENST00000374922.4 |
Ensembl |
| Affymetrix Human Clariom D Assay |
TC0X00007397.hg.1 |
ENST00000374922.4 |
Ensembl |
| Affymetrix Human Clariom D Assay |
TC0X00007397.hg.1 |
ENST00000374922.4 |
Ensembl |
| Affymetrix Human Clariom D Assay |
TC0X00007397.hg.1 |
ENST00000374922.4 |
Ensembl |
| Affymetrix Human Clariom D Assay |
TC0X00007397.hg.1 |
ENST00000374922.4 |
Ensembl |
| Affymetrix Human Clariom D Assay |
TC0X00007397.hg.1 |
ENST00000374922.4 |
Ensembl |
| Affymetrix Human Clariom D Assay |
TC0X00007397.hg.1 |
ENST00000374922.4 |
Ensembl |
| Affymetrix Human Clariom D Assay |
TC0X00007397.hg.1 |
ENST00000374922.4 |
Ensembl |
| Affymetrix Human Clariom D Assay |
TC0X00007397.hg.1 |
ENST00000374922.4 |
Ensembl |
| Affymetrix Human Clariom D Assay |
TC0X00007397.hg.1 |
ENST00000374922.4 |
Ensembl |
| Affymetrix Human Clariom D Assay |
TC0X00007397.hg.1 |
ENST00000374922.4 |
Ensembl |
| Affymetrix Human Clariom D Assay |
TC0X00007397.hg.1 |
ENST00000374922.4 |
Ensembl |
| Affymetrix Human Clariom D Assay |
TC0X00007397.hg.1 |
ENST00000423617.1 |
Ensembl |
| Affymetrix Human Clariom D Assay |
TC0X00007397.hg.1 |
ENST00000423617.1 |
Ensembl |
| Affymetrix Human Clariom D Assay |
TC0X00007397.hg.1 |
ENST00000423617.1 |
Ensembl |
| Affymetrix Human Clariom D Assay |
TC0X00007397.hg.1 |
ENST00000423617.1 |
Ensembl |
| Affymetrix Human Clariom D Assay |
TC0X00007397.hg.1 |
ENST00000423617.1 |
Ensembl |
| Affymetrix Human Clariom D Assay |
TC0X00007397.hg.1 |
ENST00000423617.1 |
Ensembl |
| Affymetrix Human Clariom D Assay |
TC0X00007397.hg.1 |
ENST00000451583.1 |
Ensembl |
| Affymetrix Human Clariom D Assay |
TC0X00007397.hg.1 |
ENST00000451583.1 |
Ensembl |
| Affymetrix Human Clariom D Assay |
TC0X00007397.hg.1 |
ENST00000451583.1 |
Ensembl |
| Affymetrix Human Clariom D Assay |
TC0X00007397.hg.1 |
ENST00000451583.1 |
Ensembl |
| Affymetrix Human Clariom D Assay |
TC0X00007397.hg.1 |
NR_015367.2 |
Refseq |
| Affymetrix Human Clariom D Assay |
TC0X00007397.hg.1 |
NR_015367.2 |
Refseq |
| Affymetrix Human Clariom D Assay |
TC0X00007397.hg.1 |
NR_015367.2 |
Refseq |
| Affymetrix Human Clariom D Assay |
TC0X00007397.hg.1 |
NR_015367.2 |
Refseq |
| Affymetrix Human Clariom D Assay |
TC0X00007397.hg.1 |
NR_015367.2 |
Refseq |
| Affymetrix Human Clariom D Assay |
TC0X00007397.hg.1 |
NR_015367.2 |
Refseq |
| Affymetrix Human Clariom D Assay |
TC0X00007397.hg.1 |
NR_015367.2 |
Refseq |
| Affymetrix Human Clariom D Assay |
TC0X00007397.hg.1 |
NR_015367.2 |
Refseq |
| Affymetrix Human Clariom D Assay |
PSR0X00150874.hg.1 |
ENST00000374922.4 |
Ensembl |
| Affymetrix Human Clariom D Assay |
PSR0X00150874.hg.1 |
ENST00000374922.4 |
Ensembl |
| Affymetrix Human Clariom D Assay |
PSR0X00150874.hg.1 |
ENST00000451583.1 |
Ensembl |
| Affymetrix Human Clariom D Assay |
PSR0X00150874.hg.1 |
NR_015367.2 |
Refseq |
| Affymetrix Human Clariom D Assay |
PSR0X00150874.hg.1 |
ENST00000374922.4 |
Ensembl |
| Affymetrix Human Clariom D Assay |
PSR0X00150874.hg.1 |
ENST00000374922.4 |
Ensembl |
| Affymetrix Human Clariom D Assay |
PSR0X00150874.hg.1 |
ENST00000451583.1 |
Ensembl |
| Affymetrix Human Clariom D Assay |
PSR0X00150874.hg.1 |
NR_015367.2 |
Refseq |
| Affymetrix Human Clariom D Assay |
PSR0X00150874.hg.1 |
TC0X00007397.hg.1 |
Clariom D TC |