| Array Name |
Probe IDs |
Targets |
Target Type |
| Affymetrix Human Transcriptome Array 2.0 |
PSR19023955.hg.1 |
ENST00000336334.2 |
Ensembl |
| Affymetrix Human Transcriptome Array 2.0 |
PSR19023955.hg.1 |
ENST00000391809.1 |
Ensembl |
| Affymetrix Human Transcriptome Array 2.0 |
PSR19023955.hg.1 |
NM_001077491.1 |
Refseq |
| Affymetrix Human Transcriptome Array 2.0 |
PSR19023955.hg.1 |
NM_001077492.1 |
Refseq |
| Affymetrix Human Transcriptome Array 2.0 |
PSR19023955.hg.1 |
NM_012427.4 |
Refseq |
| Affymetrix Human Transcriptome Array 2.0 |
PSR19023955.hg.1 |
TC19001763.hg.1 |
HTA TC |
| Affymetrix Human Transcriptome Array 2.0 |
TC1900011228.hg.1 |
ENST00000336334.7 |
Ensembl |
| Affymetrix Human Transcriptome Array 2.0 |
TC1900011228.hg.1 |
ENST00000336334.7 |
Ensembl |
| Affymetrix Human Transcriptome Array 2.0 |
TC1900011228.hg.1 |
ENST00000336334.7 |
Ensembl |
| Affymetrix Human Transcriptome Array 2.0 |
TC1900011228.hg.1 |
ENST00000336334.7 |
Ensembl |
| Affymetrix Human Transcriptome Array 2.0 |
TC1900011228.hg.1 |
ENST00000336334.7 |
Ensembl |
| Affymetrix Human Transcriptome Array 2.0 |
TC1900011228.hg.1 |
ENST00000336334.7 |
Ensembl |
| Affymetrix Human Transcriptome Array 2.0 |
TC1900011228.hg.1 |
ENST00000336334.7 |
Ensembl |
| Affymetrix Human Transcriptome Array 2.0 |
TC1900011228.hg.1 |
ENST00000336334.7 |
Ensembl |
| Affymetrix Human Transcriptome Array 2.0 |
TC1900011228.hg.1 |
ENST00000336334.7 |
Ensembl |
| Affymetrix Human Transcriptome Array 2.0 |
TC1900011228.hg.1 |
ENST00000336334.7 |
Ensembl |
| Affymetrix Human Transcriptome Array 2.0 |
TC1900011228.hg.1 |
ENST00000336334.7 |
Ensembl |
| Affymetrix Human Transcriptome Array 2.0 |
TC1900011228.hg.1 |
ENST00000391809.6 |
Ensembl |
| Affymetrix Human Transcriptome Array 2.0 |
TC1900011228.hg.1 |
ENST00000391809.6 |
Ensembl |
| Affymetrix Human Transcriptome Array 2.0 |
TC1900011228.hg.1 |
ENST00000391809.6 |
Ensembl |
| Affymetrix Human Transcriptome Array 2.0 |
TC1900011228.hg.1 |
ENST00000391809.6 |
Ensembl |
| Affymetrix Human Transcriptome Array 2.0 |
TC1900011228.hg.1 |
ENST00000391809.6 |
Ensembl |
| Affymetrix Human Transcriptome Array 2.0 |
TC1900011228.hg.1 |
ENST00000391809.6 |
Ensembl |
| Affymetrix Human Transcriptome Array 2.0 |
TC1900011228.hg.1 |
ENST00000391809.6 |
Ensembl |
| Affymetrix Human Transcriptome Array 2.0 |
TC1900011228.hg.1 |
ENST00000391809.6 |
Ensembl |
| Affymetrix Human Transcriptome Array 2.0 |
TC1900011228.hg.1 |
ENST00000391809.6 |
Ensembl |
| Affymetrix Human Transcriptome Array 2.0 |
TC1900011228.hg.1 |
ENST00000593428.5 |
Ensembl |
| Affymetrix Human Transcriptome Array 2.0 |
TC1900011228.hg.1 |
ENST00000593428.5 |
Ensembl |
| Affymetrix Human Transcriptome Array 2.0 |
TC1900011228.hg.1 |
ENST00000593428.5 |
Ensembl |
| Affymetrix Human Transcriptome Array 2.0 |
TC1900011228.hg.1 |
ENST00000593428.5 |
Ensembl |
| Affymetrix Human Transcriptome Array 2.0 |
TC1900011228.hg.1 |
ENST00000593428.5 |
Ensembl |
| Affymetrix Human Transcriptome Array 2.0 |
TC1900011228.hg.1 |
ENST00000593428.5 |
Ensembl |
| Affymetrix Human Transcriptome Array 2.0 |
TC1900011228.hg.1 |
ENST00000593428.5 |
Ensembl |
| Affymetrix Human Transcriptome Array 2.0 |
TC1900011228.hg.1 |
ENST00000593428.5 |
Ensembl |
| Affymetrix Human Transcriptome Array 2.0 |
TC1900011228.hg.1 |
ENST00000594846.1 |
Ensembl |
| Affymetrix Human Transcriptome Array 2.0 |
TC1900011228.hg.1 |
ENST00000594846.1 |
Ensembl |
| Affymetrix Human Transcriptome Array 2.0 |
TC1900011228.hg.1 |
ENST00000594846.1 |
Ensembl |
| Affymetrix Human Transcriptome Array 2.0 |
TC1900011228.hg.1 |
ENST00000594846.1 |
Ensembl |
| Affymetrix Human Transcriptome Array 2.0 |
TC1900011228.hg.1 |
ENST00000595585.1 |
Ensembl |
| Affymetrix Human Transcriptome Array 2.0 |
TC1900011228.hg.1 |
ENST00000595585.1 |
Ensembl |
| Affymetrix Human Transcriptome Array 2.0 |
TC1900011228.hg.1 |
ENST00000595585.1 |
Ensembl |
| Affymetrix Human Transcriptome Array 2.0 |
TC1900011228.hg.1 |
ENST00000595585.1 |
Ensembl |
| Affymetrix Human Transcriptome Array 2.0 |
TC1900011228.hg.1 |
ENST00000595585.1 |
Ensembl |
| Affymetrix Human Transcriptome Array 2.0 |
TC1900011228.hg.1 |
ENST00000595585.1 |
Ensembl |
| Affymetrix Human Transcriptome Array 2.0 |
TC1900011228.hg.1 |
ENST00000595585.1 |
Ensembl |
| Affymetrix Human Transcriptome Array 2.0 |
TC1900011228.hg.1 |
NM_001077491.1 |
Refseq |
| Affymetrix Human Transcriptome Array 2.0 |
TC1900011228.hg.1 |
NM_001077491.1 |
Refseq |
| Affymetrix Human Transcriptome Array 2.0 |
TC1900011228.hg.1 |
NM_001077491.1 |
Refseq |
| Affymetrix Human Transcriptome Array 2.0 |
TC1900011228.hg.1 |
NM_001077491.1 |
Refseq |
| Affymetrix Human Transcriptome Array 2.0 |
TC1900011228.hg.1 |
NM_001077491.1 |
Refseq |
| Affymetrix Human Transcriptome Array 2.0 |
TC1900011228.hg.1 |
NM_001077491.1 |
Refseq |
| Affymetrix Human Transcriptome Array 2.0 |
TC1900011228.hg.1 |
NM_001077491.1 |
Refseq |
| Affymetrix Human Transcriptome Array 2.0 |
TC1900011228.hg.1 |
NM_001077491.1 |
Refseq |
| Affymetrix Human Transcriptome Array 2.0 |
TC1900011228.hg.1 |
NM_001077491.1 |
Refseq |
| Affymetrix Human Transcriptome Array 2.0 |
TC1900011228.hg.1 |
NM_001077492.1 |
Refseq |
| Affymetrix Human Transcriptome Array 2.0 |
TC1900011228.hg.1 |
NM_001077492.1 |
Refseq |
| Affymetrix Human Transcriptome Array 2.0 |
TC1900011228.hg.1 |
NM_001077492.1 |
Refseq |
| Affymetrix Human Transcriptome Array 2.0 |
TC1900011228.hg.1 |
NM_001077492.1 |
Refseq |
| Affymetrix Human Transcriptome Array 2.0 |
TC1900011228.hg.1 |
NM_001077492.1 |
Refseq |
| Affymetrix Human Transcriptome Array 2.0 |
TC1900011228.hg.1 |
NM_001077492.1 |
Refseq |
| Affymetrix Human Transcriptome Array 2.0 |
TC1900011228.hg.1 |
NM_001077492.1 |
Refseq |
| Affymetrix Human Transcriptome Array 2.0 |
TC1900011228.hg.1 |
NM_001077492.1 |
Refseq |
| Affymetrix Human Transcriptome Array 2.0 |
TC1900011228.hg.1 |
NM_012427.4 |
Refseq |
| Affymetrix Human Transcriptome Array 2.0 |
TC1900011228.hg.1 |
NM_012427.4 |
Refseq |
| Affymetrix Human Transcriptome Array 2.0 |
TC1900011228.hg.1 |
NM_012427.4 |
Refseq |
| Affymetrix Human Transcriptome Array 2.0 |
TC1900011228.hg.1 |
NM_012427.4 |
Refseq |
| Affymetrix Human Transcriptome Array 2.0 |
TC1900011228.hg.1 |
NM_012427.4 |
Refseq |
| Affymetrix Human Transcriptome Array 2.0 |
TC1900011228.hg.1 |
NM_012427.4 |
Refseq |
| Affymetrix Human Transcriptome Array 2.0 |
TC1900011228.hg.1 |
NM_012427.4 |
Refseq |
| Affymetrix Human Transcriptome Array 2.0 |
TC1900011228.hg.1 |
NM_012427.4 |
Refseq |
| Affymetrix Human Transcriptome Array 2.0 |
TC1900011228.hg.1 |
NM_012427.4 |
Refseq |
| Affymetrix Human Transcriptome Array 2.0 |
TC1900011228.hg.1 |
NM_012427.4 |
Refseq |
| Affymetrix Human Transcriptome Array 2.0 |
TC1900011228.hg.1 |
NM_012427.4 |
Refseq |
| Affymetrix Human Clariom D Assay |
PSR1900194388.hg.1 |
ENST00000336334.7 |
Ensembl |
| Affymetrix Human Clariom D Assay |
PSR1900194388.hg.1 |
ENST00000391809.6 |
Ensembl |
| Affymetrix Human Clariom D Assay |
PSR1900194388.hg.1 |
ENST00000593428.5 |
Ensembl |
| Affymetrix Human Clariom D Assay |
PSR1900194388.hg.1 |
ENST00000594846.1 |
Ensembl |
| Affymetrix Human Clariom D Assay |
PSR1900194388.hg.1 |
ENST00000595585.1 |
Ensembl |
| Affymetrix Human Clariom D Assay |
PSR1900194388.hg.1 |
NM_001077491.1 |
Refseq |
| Affymetrix Human Clariom D Assay |
PSR1900194388.hg.1 |
NM_001077492.1 |
Refseq |
| Affymetrix Human Clariom D Assay |
PSR1900194388.hg.1 |
NM_012427.4 |
Refseq |
| Affymetrix Human Clariom D Assay |
PSR1900194388.hg.1 |
TC1900011228.hg.1 |
Clariom D TC |
| Affymetrix Human Clariom D Assay |
PSR1900194388.hg.1 |
ENST00000336334.7 |
Ensembl |
| Affymetrix Human Clariom D Assay |
PSR1900194388.hg.1 |
ENST00000391809.6 |
Ensembl |
| Affymetrix Human Clariom D Assay |
PSR1900194388.hg.1 |
ENST00000593428.5 |
Ensembl |
| Affymetrix Human Clariom D Assay |
PSR1900194388.hg.1 |
ENST00000594846.1 |
Ensembl |
| Affymetrix Human Clariom D Assay |
PSR1900194388.hg.1 |
ENST00000595585.1 |
Ensembl |
| Affymetrix Human Clariom D Assay |
PSR1900194388.hg.1 |
NM_001077491.1 |
Refseq |
| Affymetrix Human Clariom D Assay |
PSR1900194388.hg.1 |
NM_001077492.1 |
Refseq |
| Affymetrix Human Clariom D Assay |
PSR1900194388.hg.1 |
NM_012427.4 |
Refseq |
| Affymetrix Human Clariom D Assay |
TC1900011228.hg.1 |
ENST00000336334.7 |
Ensembl |
| Affymetrix Human Clariom D Assay |
TC1900011228.hg.1 |
ENST00000336334.7 |
Ensembl |
| Affymetrix Human Clariom D Assay |
TC1900011228.hg.1 |
ENST00000336334.7 |
Ensembl |
| Affymetrix Human Clariom D Assay |
TC1900011228.hg.1 |
ENST00000336334.7 |
Ensembl |
| Affymetrix Human Clariom D Assay |
TC1900011228.hg.1 |
ENST00000336334.7 |
Ensembl |
| Affymetrix Human Clariom D Assay |
TC1900011228.hg.1 |
ENST00000336334.7 |
Ensembl |
| Affymetrix Human Clariom D Assay |
TC1900011228.hg.1 |
ENST00000336334.7 |
Ensembl |
| Affymetrix Human Clariom D Assay |
TC1900011228.hg.1 |
ENST00000336334.7 |
Ensembl |
| Affymetrix Human Clariom D Assay |
TC1900011228.hg.1 |
ENST00000336334.7 |
Ensembl |
| Affymetrix Human Clariom D Assay |
TC1900011228.hg.1 |
ENST00000336334.7 |
Ensembl |
| Affymetrix Human Clariom D Assay |
TC1900011228.hg.1 |
ENST00000336334.7 |
Ensembl |
| Affymetrix Human Clariom D Assay |
TC1900011228.hg.1 |
ENST00000391809.6 |
Ensembl |
| Affymetrix Human Clariom D Assay |
TC1900011228.hg.1 |
ENST00000391809.6 |
Ensembl |
| Affymetrix Human Clariom D Assay |
TC1900011228.hg.1 |
ENST00000391809.6 |
Ensembl |
| Affymetrix Human Clariom D Assay |
TC1900011228.hg.1 |
ENST00000391809.6 |
Ensembl |
| Affymetrix Human Clariom D Assay |
TC1900011228.hg.1 |
ENST00000391809.6 |
Ensembl |
| Affymetrix Human Clariom D Assay |
TC1900011228.hg.1 |
ENST00000391809.6 |
Ensembl |
| Affymetrix Human Clariom D Assay |
TC1900011228.hg.1 |
ENST00000391809.6 |
Ensembl |
| Affymetrix Human Clariom D Assay |
TC1900011228.hg.1 |
ENST00000391809.6 |
Ensembl |
| Affymetrix Human Clariom D Assay |
TC1900011228.hg.1 |
ENST00000391809.6 |
Ensembl |
| Affymetrix Human Clariom D Assay |
TC1900011228.hg.1 |
ENST00000593428.5 |
Ensembl |
| Affymetrix Human Clariom D Assay |
TC1900011228.hg.1 |
ENST00000593428.5 |
Ensembl |
| Affymetrix Human Clariom D Assay |
TC1900011228.hg.1 |
ENST00000593428.5 |
Ensembl |
| Affymetrix Human Clariom D Assay |
TC1900011228.hg.1 |
ENST00000593428.5 |
Ensembl |
| Affymetrix Human Clariom D Assay |
TC1900011228.hg.1 |
ENST00000593428.5 |
Ensembl |
| Affymetrix Human Clariom D Assay |
TC1900011228.hg.1 |
ENST00000593428.5 |
Ensembl |
| Affymetrix Human Clariom D Assay |
TC1900011228.hg.1 |
ENST00000593428.5 |
Ensembl |
| Affymetrix Human Clariom D Assay |
TC1900011228.hg.1 |
ENST00000593428.5 |
Ensembl |
| Affymetrix Human Clariom D Assay |
TC1900011228.hg.1 |
ENST00000594846.1 |
Ensembl |
| Affymetrix Human Clariom D Assay |
TC1900011228.hg.1 |
ENST00000594846.1 |
Ensembl |
| Affymetrix Human Clariom D Assay |
TC1900011228.hg.1 |
ENST00000594846.1 |
Ensembl |
| Affymetrix Human Clariom D Assay |
TC1900011228.hg.1 |
ENST00000594846.1 |
Ensembl |
| Affymetrix Human Clariom D Assay |
TC1900011228.hg.1 |
ENST00000595585.1 |
Ensembl |
| Affymetrix Human Clariom D Assay |
TC1900011228.hg.1 |
ENST00000595585.1 |
Ensembl |
| Affymetrix Human Clariom D Assay |
TC1900011228.hg.1 |
ENST00000595585.1 |
Ensembl |
| Affymetrix Human Clariom D Assay |
TC1900011228.hg.1 |
ENST00000595585.1 |
Ensembl |
| Affymetrix Human Clariom D Assay |
TC1900011228.hg.1 |
ENST00000595585.1 |
Ensembl |
| Affymetrix Human Clariom D Assay |
TC1900011228.hg.1 |
ENST00000595585.1 |
Ensembl |
| Affymetrix Human Clariom D Assay |
TC1900011228.hg.1 |
ENST00000595585.1 |
Ensembl |
| Affymetrix Human Clariom D Assay |
TC1900011228.hg.1 |
NM_001077491.1 |
Refseq |
| Affymetrix Human Clariom D Assay |
TC1900011228.hg.1 |
NM_001077491.1 |
Refseq |
| Affymetrix Human Clariom D Assay |
TC1900011228.hg.1 |
NM_001077491.1 |
Refseq |
| Affymetrix Human Clariom D Assay |
TC1900011228.hg.1 |
NM_001077491.1 |
Refseq |
| Affymetrix Human Clariom D Assay |
TC1900011228.hg.1 |
NM_001077491.1 |
Refseq |
| Affymetrix Human Clariom D Assay |
TC1900011228.hg.1 |
NM_001077491.1 |
Refseq |
| Affymetrix Human Clariom D Assay |
TC1900011228.hg.1 |
NM_001077491.1 |
Refseq |
| Affymetrix Human Clariom D Assay |
TC1900011228.hg.1 |
NM_001077491.1 |
Refseq |
| Affymetrix Human Clariom D Assay |
TC1900011228.hg.1 |
NM_001077491.1 |
Refseq |
| Affymetrix Human Clariom D Assay |
TC1900011228.hg.1 |
NM_001077492.1 |
Refseq |
| Affymetrix Human Clariom D Assay |
TC1900011228.hg.1 |
NM_001077492.1 |
Refseq |
| Affymetrix Human Clariom D Assay |
TC1900011228.hg.1 |
NM_001077492.1 |
Refseq |
| Affymetrix Human Clariom D Assay |
TC1900011228.hg.1 |
NM_001077492.1 |
Refseq |
| Affymetrix Human Clariom D Assay |
TC1900011228.hg.1 |
NM_001077492.1 |
Refseq |
| Affymetrix Human Clariom D Assay |
TC1900011228.hg.1 |
NM_001077492.1 |
Refseq |
| Affymetrix Human Clariom D Assay |
TC1900011228.hg.1 |
NM_001077492.1 |
Refseq |
| Affymetrix Human Clariom D Assay |
TC1900011228.hg.1 |
NM_001077492.1 |
Refseq |
| Affymetrix Human Clariom D Assay |
TC1900011228.hg.1 |
NM_012427.4 |
Refseq |
| Affymetrix Human Clariom D Assay |
TC1900011228.hg.1 |
NM_012427.4 |
Refseq |
| Affymetrix Human Clariom D Assay |
TC1900011228.hg.1 |
NM_012427.4 |
Refseq |
| Affymetrix Human Clariom D Assay |
TC1900011228.hg.1 |
NM_012427.4 |
Refseq |
| Affymetrix Human Clariom D Assay |
TC1900011228.hg.1 |
NM_012427.4 |
Refseq |
| Affymetrix Human Clariom D Assay |
TC1900011228.hg.1 |
NM_012427.4 |
Refseq |
| Affymetrix Human Clariom D Assay |
TC1900011228.hg.1 |
NM_012427.4 |
Refseq |
| Affymetrix Human Clariom D Assay |
TC1900011228.hg.1 |
NM_012427.4 |
Refseq |
| Affymetrix Human Clariom D Assay |
TC1900011228.hg.1 |
NM_012427.4 |
Refseq |
| Affymetrix Human Clariom D Assay |
TC1900011228.hg.1 |
NM_012427.4 |
Refseq |
| Affymetrix Human Clariom D Assay |
TC1900011228.hg.1 |
NM_012427.4 |
Refseq |