Assay Details
| Gene Symbol: ASB14 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | NM_001142733.2 | NP_001136205.2 | 2 - 3 | 246 | 131 | |
| XM_017005736.1 | XP_016861225.1 | 2 - 3 | 596 | 131 | ||
| XM_017005737.1 | XP_016861226.1 | 2 - 3 | 560 | 131 | ||
| GenBank mRNA | AL832472.1 | - | 2 - 3 | 247 | 131 | |
More Information
Updated Mapping Information
This assay targets the same location as HTA transcript cluster(s) TC03001491.hg.1
Set Membership: |
|
Panther Classification:
Gene Ontology Categories:
MicroArray Probes:
Affymetrix
| Array Name | Probe IDs | Targets | Target Type |
|---|---|---|---|
| Affymetrix Human Transcriptome Array 2.0 | PSR03027131.hg.1 | TC03001491.hg.1 | HTA TC |
| Affymetrix Human Transcriptome Array 2.0 | PSR03027131.hg.1 | ENST00000389601.3 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | PSR03027131.hg.1 | ENST00000438870.2 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | PSR03027131.hg.1 | ENST00000487349.1 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | PSR03027131.hg.1 | NM_001142733.2 | Refseq |
| Affymetrix Human Transcriptome Array 2.0 | PSR03027126.hg.1 | TC03001491.hg.1 | HTA TC |
| Affymetrix Human Transcriptome Array 2.0 | PSR03027126.hg.1 | ENST00000389601.3 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | PSR03027126.hg.1 | ENST00000438870.2 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | PSR03027126.hg.1 | ENST00000487349.1 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | PSR03027126.hg.1 | NM_001142733.2 | Refseq |
| Affymetrix Human Transcriptome Array 2.0 | PSR03027130.hg.1 | TC03001491.hg.1 | HTA TC |
| Affymetrix Human Transcriptome Array 2.0 | PSR03027130.hg.1 | ENST00000389601.3 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | PSR03027130.hg.1 | ENST00000438870.2 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | PSR03027130.hg.1 | ENST00000487349.1 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | PSR03027130.hg.1 | NM_001142733.2 | Refseq |
| Affymetrix Human Transcriptome Array 2.0 | TC0300011266.hg.1 | ENST00000389601.3 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0300011266.hg.1 | ENST00000389601.3 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0300011266.hg.1 | ENST00000389601.3 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0300011266.hg.1 | ENST00000389601.3 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0300011266.hg.1 | ENST00000389601.3 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0300011266.hg.1 | ENST00000389601.3 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0300011266.hg.1 | ENST00000389601.3 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0300011266.hg.1 | ENST00000389601.3 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0300011266.hg.1 | ENST00000389601.3 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0300011266.hg.1 | ENST00000389601.3 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0300011266.hg.1 | ENST00000389601.3 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0300011266.hg.1 | ENST00000389601.3 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0300011266.hg.1 | ENST00000389601.3 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0300011266.hg.1 | ENST00000389601.3 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0300011266.hg.1 | ENST00000487349.5 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0300011266.hg.1 | ENST00000487349.5 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0300011266.hg.1 | ENST00000487349.5 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0300011266.hg.1 | ENST00000487349.5 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0300011266.hg.1 | ENST00000487349.5 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0300011266.hg.1 | ENST00000487349.5 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0300011266.hg.1 | ENST00000487349.5 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0300011266.hg.1 | ENST00000487349.5 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0300011266.hg.1 | ENST00000487349.5 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0300011266.hg.1 | ENST00000487349.5 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0300011266.hg.1 | ENST00000487349.5 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0300011266.hg.1 | ENST00000487349.5 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0300011266.hg.1 | ENST00000487349.5 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0300011266.hg.1 | ENST00000487349.5 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0300011266.hg.1 | ENST00000515033.1 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0300011266.hg.1 | ENST00000515033.1 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0300011266.hg.1 | ENST00000515033.1 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0300011266.hg.1 | ENST00000515033.1 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0300011266.hg.1 | ENST00000515033.1 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0300011266.hg.1 | NM_001142733.2 | Refseq |
| Affymetrix Human Transcriptome Array 2.0 | TC0300011266.hg.1 | NM_001142733.2 | Refseq |
| Affymetrix Human Transcriptome Array 2.0 | TC0300011266.hg.1 | NM_001142733.2 | Refseq |
| Affymetrix Human Transcriptome Array 2.0 | TC0300011266.hg.1 | NM_001142733.2 | Refseq |
| Affymetrix Human Transcriptome Array 2.0 | TC0300011266.hg.1 | NM_001142733.2 | Refseq |
| Affymetrix Human Transcriptome Array 2.0 | TC0300011266.hg.1 | NM_001142733.2 | Refseq |
| Affymetrix Human Transcriptome Array 2.0 | TC0300011266.hg.1 | NM_001142733.2 | Refseq |
| Affymetrix Human Transcriptome Array 2.0 | TC0300011266.hg.1 | NM_001142733.2 | Refseq |
| Affymetrix Human Transcriptome Array 2.0 | TC0300011266.hg.1 | NM_001142733.2 | Refseq |
| Affymetrix Human Transcriptome Array 2.0 | TC0300011266.hg.1 | NM_001142733.2 | Refseq |
| Affymetrix Human Transcriptome Array 2.0 | TC0300011266.hg.1 | NM_001142733.2 | Refseq |
| Affymetrix Human Transcriptome Array 2.0 | TC0300011266.hg.1 | NM_001142733.2 | Refseq |
| Affymetrix Human Transcriptome Array 2.0 | TC0300011266.hg.1 | NM_001142733.2 | Refseq |
| Affymetrix Human Transcriptome Array 2.0 | TC0300011266.hg.1 | NM_001142733.2 | Refseq |
| Affymetrix Human Transcriptome Array 2.0 | TC0300011266.hg.1 | NM_130387.5 | Refseq |
| Affymetrix Human Transcriptome Array 2.0 | TC0300011266.hg.1 | NM_130387.5 | Refseq |
| Affymetrix Human Transcriptome Array 2.0 | TC0300011266.hg.1 | NM_130387.5 | Refseq |
| Affymetrix Human Transcriptome Array 2.0 | TC0300011266.hg.1 | NM_130387.5 | Refseq |
| Affymetrix Human Transcriptome Array 2.0 | TC0300011266.hg.1 | NM_130387.5 | Refseq |
| Affymetrix Human Transcriptome Array 2.0 | TC0300011266.hg.1 | NM_130387.5 | Refseq |
| Affymetrix Human Transcriptome Array 2.0 | TC0300011266.hg.1 | NM_130387.5 | Refseq |
| Affymetrix Human Transcriptome Array 2.0 | PSR03027129.hg.1 | TC03001491.hg.1 | HTA TC |
| Affymetrix Human Transcriptome Array 2.0 | PSR03027129.hg.1 | ENST00000389601.3 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | PSR03027129.hg.1 | ENST00000438870.2 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | PSR03027129.hg.1 | ENST00000487349.1 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | PSR03027129.hg.1 | NM_001142733.2 | Refseq |
| Affymetrix Human Clariom D Assay | PSR0300184463.hg.1 | ENST00000389601.3 | Ensembl |
| Affymetrix Human Clariom D Assay | PSR0300184463.hg.1 | ENST00000487349.5 | Ensembl |
| Affymetrix Human Clariom D Assay | PSR0300184463.hg.1 | NM_001142733.2 | Refseq |
| Affymetrix Human Clariom D Assay | PSR0300184463.hg.1 | ENST00000389601.3 | Ensembl |
| Affymetrix Human Clariom D Assay | PSR0300184463.hg.1 | ENST00000487349.5 | Ensembl |
| Affymetrix Human Clariom D Assay | PSR0300184463.hg.1 | NM_001142733.2 | Refseq |
| Affymetrix Human Clariom D Assay | PSR0300184463.hg.1 | TC0300011266.hg.1 | Clariom D TC |
| Affymetrix Human Clariom D Assay | PSR0300184469.hg.1 | ENST00000389601.3 | Ensembl |
| Affymetrix Human Clariom D Assay | PSR0300184469.hg.1 | ENST00000487349.5 | Ensembl |
| Affymetrix Human Clariom D Assay | PSR0300184469.hg.1 | NM_001142733.2 | Refseq |
| Affymetrix Human Clariom D Assay | PSR0300184469.hg.1 | ENST00000389601.3 | Ensembl |
| Affymetrix Human Clariom D Assay | PSR0300184469.hg.1 | ENST00000487349.5 | Ensembl |
| Affymetrix Human Clariom D Assay | PSR0300184469.hg.1 | NM_001142733.2 | Refseq |
| Affymetrix Human Clariom D Assay | PSR0300184469.hg.1 | TC0300011266.hg.1 | Clariom D TC |
| Affymetrix Human Clariom D Assay | PSR0300184468.hg.1 | ENST00000389601.3 | Ensembl |
| Affymetrix Human Clariom D Assay | PSR0300184468.hg.1 | ENST00000487349.5 | Ensembl |
| Affymetrix Human Clariom D Assay | PSR0300184468.hg.1 | NM_001142733.2 | Refseq |
| Affymetrix Human Clariom D Assay | PSR0300184468.hg.1 | ENST00000389601.3 | Ensembl |
| Affymetrix Human Clariom D Assay | PSR0300184468.hg.1 | ENST00000487349.5 | Ensembl |
| Affymetrix Human Clariom D Assay | PSR0300184468.hg.1 | NM_001142733.2 | Refseq |
| Affymetrix Human Clariom D Assay | PSR0300184468.hg.1 | TC0300011266.hg.1 | Clariom D TC |
| Affymetrix Human Clariom D Assay | PSR0300184466.hg.1 | ENST00000389601.3 | Ensembl |
| Affymetrix Human Clariom D Assay | PSR0300184466.hg.1 | ENST00000487349.5 | Ensembl |
| Affymetrix Human Clariom D Assay | PSR0300184466.hg.1 | NM_001142733.2 | Refseq |
| Affymetrix Human Clariom D Assay | PSR0300184466.hg.1 | ENST00000389601.3 | Ensembl |
| Affymetrix Human Clariom D Assay | PSR0300184466.hg.1 | ENST00000487349.5 | Ensembl |
| Affymetrix Human Clariom D Assay | PSR0300184466.hg.1 | NM_001142733.2 | Refseq |
| Affymetrix Human Clariom D Assay | PSR0300184466.hg.1 | TC0300011266.hg.1 | Clariom D TC |
| Affymetrix Human Clariom D Assay | PSR0300184467.hg.1 | ENST00000389601.3 | Ensembl |
| Affymetrix Human Clariom D Assay | PSR0300184467.hg.1 | ENST00000487349.5 | Ensembl |
| Affymetrix Human Clariom D Assay | PSR0300184467.hg.1 | NM_001142733.2 | Refseq |
| Affymetrix Human Clariom D Assay | PSR0300184467.hg.1 | ENST00000389601.3 | Ensembl |
| Affymetrix Human Clariom D Assay | PSR0300184467.hg.1 | ENST00000487349.5 | Ensembl |
| Affymetrix Human Clariom D Assay | PSR0300184467.hg.1 | NM_001142733.2 | Refseq |
| Affymetrix Human Clariom D Assay | PSR0300184467.hg.1 | TC0300011266.hg.1 | Clariom D TC |
| Affymetrix Human Clariom D Assay | TC0300011266.hg.1 | ENST00000389601.3 | Ensembl |
| Affymetrix Human Clariom D Assay | TC0300011266.hg.1 | ENST00000389601.3 | Ensembl |
| Affymetrix Human Clariom D Assay | TC0300011266.hg.1 | ENST00000389601.3 | Ensembl |
| Affymetrix Human Clariom D Assay | TC0300011266.hg.1 | ENST00000389601.3 | Ensembl |
| Affymetrix Human Clariom D Assay | TC0300011266.hg.1 | ENST00000389601.3 | Ensembl |
| Affymetrix Human Clariom D Assay | TC0300011266.hg.1 | ENST00000389601.3 | Ensembl |
| Affymetrix Human Clariom D Assay | TC0300011266.hg.1 | ENST00000389601.3 | Ensembl |
| Affymetrix Human Clariom D Assay | TC0300011266.hg.1 | ENST00000389601.3 | Ensembl |
| Affymetrix Human Clariom D Assay | TC0300011266.hg.1 | ENST00000389601.3 | Ensembl |
| Affymetrix Human Clariom D Assay | TC0300011266.hg.1 | ENST00000389601.3 | Ensembl |
| Affymetrix Human Clariom D Assay | TC0300011266.hg.1 | ENST00000389601.3 | Ensembl |
| Affymetrix Human Clariom D Assay | TC0300011266.hg.1 | ENST00000389601.3 | Ensembl |
| Affymetrix Human Clariom D Assay | TC0300011266.hg.1 | ENST00000389601.3 | Ensembl |
| Affymetrix Human Clariom D Assay | TC0300011266.hg.1 | ENST00000389601.3 | Ensembl |
| Affymetrix Human Clariom D Assay | TC0300011266.hg.1 | ENST00000487349.5 | Ensembl |
| Affymetrix Human Clariom D Assay | TC0300011266.hg.1 | ENST00000487349.5 | Ensembl |
| Affymetrix Human Clariom D Assay | TC0300011266.hg.1 | ENST00000487349.5 | Ensembl |
| Affymetrix Human Clariom D Assay | TC0300011266.hg.1 | ENST00000487349.5 | Ensembl |
| Affymetrix Human Clariom D Assay | TC0300011266.hg.1 | ENST00000487349.5 | Ensembl |
| Affymetrix Human Clariom D Assay | TC0300011266.hg.1 | ENST00000487349.5 | Ensembl |
| Affymetrix Human Clariom D Assay | TC0300011266.hg.1 | ENST00000487349.5 | Ensembl |
| Affymetrix Human Clariom D Assay | TC0300011266.hg.1 | ENST00000487349.5 | Ensembl |
| Affymetrix Human Clariom D Assay | TC0300011266.hg.1 | ENST00000487349.5 | Ensembl |
| Affymetrix Human Clariom D Assay | TC0300011266.hg.1 | ENST00000487349.5 | Ensembl |
| Affymetrix Human Clariom D Assay | TC0300011266.hg.1 | ENST00000487349.5 | Ensembl |
| Affymetrix Human Clariom D Assay | TC0300011266.hg.1 | ENST00000487349.5 | Ensembl |
| Affymetrix Human Clariom D Assay | TC0300011266.hg.1 | ENST00000487349.5 | Ensembl |
| Affymetrix Human Clariom D Assay | TC0300011266.hg.1 | ENST00000487349.5 | Ensembl |
| Affymetrix Human Clariom D Assay | TC0300011266.hg.1 | ENST00000515033.1 | Ensembl |
| Affymetrix Human Clariom D Assay | TC0300011266.hg.1 | ENST00000515033.1 | Ensembl |
| Affymetrix Human Clariom D Assay | TC0300011266.hg.1 | ENST00000515033.1 | Ensembl |
| Affymetrix Human Clariom D Assay | TC0300011266.hg.1 | ENST00000515033.1 | Ensembl |
| Affymetrix Human Clariom D Assay | TC0300011266.hg.1 | ENST00000515033.1 | Ensembl |
| Affymetrix Human Clariom D Assay | TC0300011266.hg.1 | NM_001142733.2 | Refseq |
| Affymetrix Human Clariom D Assay | TC0300011266.hg.1 | NM_001142733.2 | Refseq |
| Affymetrix Human Clariom D Assay | TC0300011266.hg.1 | NM_001142733.2 | Refseq |
| Affymetrix Human Clariom D Assay | TC0300011266.hg.1 | NM_001142733.2 | Refseq |
| Affymetrix Human Clariom D Assay | TC0300011266.hg.1 | NM_001142733.2 | Refseq |
| Affymetrix Human Clariom D Assay | TC0300011266.hg.1 | NM_001142733.2 | Refseq |
| Affymetrix Human Clariom D Assay | TC0300011266.hg.1 | NM_001142733.2 | Refseq |
| Affymetrix Human Clariom D Assay | TC0300011266.hg.1 | NM_001142733.2 | Refseq |
| Affymetrix Human Clariom D Assay | TC0300011266.hg.1 | NM_001142733.2 | Refseq |
| Affymetrix Human Clariom D Assay | TC0300011266.hg.1 | NM_001142733.2 | Refseq |
| Affymetrix Human Clariom D Assay | TC0300011266.hg.1 | NM_001142733.2 | Refseq |
| Affymetrix Human Clariom D Assay | TC0300011266.hg.1 | NM_001142733.2 | Refseq |
| Affymetrix Human Clariom D Assay | TC0300011266.hg.1 | NM_001142733.2 | Refseq |
| Affymetrix Human Clariom D Assay | TC0300011266.hg.1 | NM_001142733.2 | Refseq |
| Affymetrix Human Clariom D Assay | TC0300011266.hg.1 | NM_130387.5 | Refseq |
| Affymetrix Human Clariom D Assay | TC0300011266.hg.1 | NM_130387.5 | Refseq |
| Affymetrix Human Clariom D Assay | TC0300011266.hg.1 | NM_130387.5 | Refseq |
| Affymetrix Human Clariom D Assay | TC0300011266.hg.1 | NM_130387.5 | Refseq |
| Affymetrix Human Clariom D Assay | TC0300011266.hg.1 | NM_130387.5 | Refseq |
| Affymetrix Human Clariom D Assay | TC0300011266.hg.1 | NM_130387.5 | Refseq |
| Affymetrix Human Clariom D Assay | TC0300011266.hg.1 | NM_130387.5 | Refseq |

ASB14
Genomic Map