Assay Details
| Gene Symbol: LGR4 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | NM_018490.2 | NP_060960.2 | 9 - 10 | 1390 | 141 | |
| GenBank mRNA | AB209743.1 | - | 9 - 10 | 1309 | 141 | |
| AF257182.1 | - | 9 - 10 | 1349 | 141 | ||
| BC033039.2 | - | 8 - 9 | 1306 | 5260923 | 141 | |
More Information
Updated Mapping Information
This assay targets the same location as HTA transcript cluster(s) TC11001509.hg.1
Set Membership: |
|
Matching Ambion siRNAs:
Gene Ontology Categories:
Function(s)
Process(es)
osteoblast differentiation
hair follicle development
G-protein coupled receptor signaling pathway
spermatogenesis
bone mineralization
male genitalia development
circadian regulation of gene expression
negative regulation of toll-like receptor signaling pathway
intestinal stem cell homeostasis
innate immune response
negative regulation of transcription, DNA-templated
positive regulation of transcription, DNA-templated
bone remodeling
digestive tract development
negative regulation of cytokine secretion
canonical Wnt signaling pathway involved in metanephric kidney development
cell differentiation involved in metanephros development
metanephric glomerulus development
metanephric nephron tubule morphogenesis
positive regulation of branching involved in ureteric bud morphogenesis
positive regulation of canonical Wnt signaling pathway
epithelial cell proliferation involved in renal tubule morphogenesis
transmembrane signaling receptor activity
G-protein coupled receptor activity
protein binding
osteoblast differentiation
hair follicle development
G-protein coupled receptor signaling pathway
spermatogenesis
bone mineralization
male genitalia development
circadian regulation of gene expression
negative regulation of toll-like receptor signaling pathway
intestinal stem cell homeostasis
innate immune response
negative regulation of transcription, DNA-templated
positive regulation of transcription, DNA-templated
bone remodeling
digestive tract development
negative regulation of cytokine secretion
canonical Wnt signaling pathway involved in metanephric kidney development
cell differentiation involved in metanephros development
metanephric glomerulus development
metanephric nephron tubule morphogenesis
positive regulation of branching involved in ureteric bud morphogenesis
positive regulation of canonical Wnt signaling pathway
epithelial cell proliferation involved in renal tubule morphogenesis
transmembrane signaling receptor activity
G-protein coupled receptor activity
protein binding
MicroArray Probes:
Affymetrix
| Array Name | Probe IDs | Targets | Target Type |
|---|---|---|---|
| Affymetrix Human Transcriptome Array 2.0 | PSR11018896.hg.1 | ENST00000379214.4 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | PSR11018896.hg.1 | ENST00000389858.4 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | PSR11018896.hg.1 | NM_018490.3 | Refseq |
| Affymetrix Human Transcriptome Array 2.0 | PSR11018896.hg.1 | TC11001509.hg.1 | HTA TC |
| Affymetrix Human Transcriptome Array 2.0 | TC1100010400.hg.1 | ENST00000379214.8 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC1100010400.hg.1 | ENST00000379214.8 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC1100010400.hg.1 | ENST00000379214.8 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC1100010400.hg.1 | ENST00000379214.8 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC1100010400.hg.1 | ENST00000379214.8 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC1100010400.hg.1 | ENST00000379214.8 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC1100010400.hg.1 | ENST00000379214.8 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC1100010400.hg.1 | ENST00000379214.8 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC1100010400.hg.1 | ENST00000379214.8 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC1100010400.hg.1 | ENST00000379214.8 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC1100010400.hg.1 | ENST00000379214.8 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC1100010400.hg.1 | ENST00000379214.8 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC1100010400.hg.1 | ENST00000379214.8 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC1100010400.hg.1 | ENST00000379214.8 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC1100010400.hg.1 | ENST00000379214.8 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC1100010400.hg.1 | ENST00000379214.8 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC1100010400.hg.1 | ENST00000379214.8 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC1100010400.hg.1 | ENST00000379214.8 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC1100010400.hg.1 | ENST00000379214.8 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC1100010400.hg.1 | ENST00000379214.8 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC1100010400.hg.1 | ENST00000379214.8 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC1100010400.hg.1 | ENST00000379214.8 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC1100010400.hg.1 | ENST00000389858.4 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC1100010400.hg.1 | ENST00000389858.4 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC1100010400.hg.1 | ENST00000389858.4 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC1100010400.hg.1 | ENST00000389858.4 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC1100010400.hg.1 | ENST00000389858.4 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC1100010400.hg.1 | ENST00000389858.4 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC1100010400.hg.1 | ENST00000389858.4 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC1100010400.hg.1 | ENST00000389858.4 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC1100010400.hg.1 | ENST00000389858.4 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC1100010400.hg.1 | ENST00000389858.4 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC1100010400.hg.1 | ENST00000389858.4 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC1100010400.hg.1 | ENST00000389858.4 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC1100010400.hg.1 | ENST00000389858.4 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC1100010400.hg.1 | ENST00000389858.4 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC1100010400.hg.1 | ENST00000389858.4 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC1100010400.hg.1 | ENST00000389858.4 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC1100010400.hg.1 | ENST00000389858.4 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC1100010400.hg.1 | ENST00000389858.4 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC1100010400.hg.1 | ENST00000389858.4 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC1100010400.hg.1 | ENST00000389858.4 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC1100010400.hg.1 | ENST00000389858.4 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC1100010400.hg.1 | ENST00000389858.4 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC1100010400.hg.1 | ENST00000480977.2 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC1100010400.hg.1 | ENST00000480977.2 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC1100010400.hg.1 | ENST00000480977.2 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC1100010400.hg.1 | ENST00000489910.1 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC1100010400.hg.1 | ENST00000489910.1 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC1100010400.hg.1 | ENST00000489910.1 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC1100010400.hg.1 | ENST00000527773.1 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC1100010400.hg.1 | ENST00000527773.1 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC1100010400.hg.1 | ENST00000527773.1 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC1100010400.hg.1 | ENST00000527773.1 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC1100010400.hg.1 | ENST00000530658.1 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC1100010400.hg.1 | ENST00000530658.1 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC1100010400.hg.1 | ENST00000530658.1 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC1100010400.hg.1 | NM_018490.3 | Refseq |
| Affymetrix Human Transcriptome Array 2.0 | TC1100010400.hg.1 | NM_018490.3 | Refseq |
| Affymetrix Human Transcriptome Array 2.0 | TC1100010400.hg.1 | NM_018490.3 | Refseq |
| Affymetrix Human Transcriptome Array 2.0 | TC1100010400.hg.1 | NM_018490.3 | Refseq |
| Affymetrix Human Transcriptome Array 2.0 | TC1100010400.hg.1 | NM_018490.3 | Refseq |
| Affymetrix Human Transcriptome Array 2.0 | TC1100010400.hg.1 | NM_018490.3 | Refseq |
| Affymetrix Human Transcriptome Array 2.0 | TC1100010400.hg.1 | NM_018490.3 | Refseq |
| Affymetrix Human Transcriptome Array 2.0 | TC1100010400.hg.1 | NM_018490.3 | Refseq |
| Affymetrix Human Transcriptome Array 2.0 | TC1100010400.hg.1 | NM_018490.3 | Refseq |
| Affymetrix Human Transcriptome Array 2.0 | TC1100010400.hg.1 | NM_018490.3 | Refseq |
| Affymetrix Human Transcriptome Array 2.0 | TC1100010400.hg.1 | NM_018490.3 | Refseq |
| Affymetrix Human Transcriptome Array 2.0 | TC1100010400.hg.1 | NM_018490.3 | Refseq |
| Affymetrix Human Transcriptome Array 2.0 | TC1100010400.hg.1 | NM_018490.3 | Refseq |
| Affymetrix Human Transcriptome Array 2.0 | TC1100010400.hg.1 | NM_018490.3 | Refseq |
| Affymetrix Human Transcriptome Array 2.0 | TC1100010400.hg.1 | NM_018490.3 | Refseq |
| Affymetrix Human Transcriptome Array 2.0 | TC1100010400.hg.1 | NM_018490.3 | Refseq |
| Affymetrix Human Transcriptome Array 2.0 | TC1100010400.hg.1 | NM_018490.3 | Refseq |
| Affymetrix Human Transcriptome Array 2.0 | TC1100010400.hg.1 | NM_018490.3 | Refseq |
| Affymetrix Human Transcriptome Array 2.0 | TC1100010400.hg.1 | NM_018490.3 | Refseq |
| Affymetrix Human Transcriptome Array 2.0 | TC1100010400.hg.1 | NM_018490.3 | Refseq |
| Affymetrix Human Transcriptome Array 2.0 | TC1100010400.hg.1 | NM_018490.3 | Refseq |
| Affymetrix Human Transcriptome Array 2.0 | TC1100010400.hg.1 | NM_018490.3 | Refseq |
| Affymetrix Human Transcriptome Array 2.0 | TC1100010400.hg.1 | NM_018490.3 | Refseq |
| Affymetrix Human Clariom D Assay | TC1100010400.hg.1 | ENST00000379214.8 | Ensembl |
| Affymetrix Human Clariom D Assay | TC1100010400.hg.1 | ENST00000379214.8 | Ensembl |
| Affymetrix Human Clariom D Assay | TC1100010400.hg.1 | ENST00000379214.8 | Ensembl |
| Affymetrix Human Clariom D Assay | TC1100010400.hg.1 | ENST00000379214.8 | Ensembl |
| Affymetrix Human Clariom D Assay | TC1100010400.hg.1 | ENST00000379214.8 | Ensembl |
| Affymetrix Human Clariom D Assay | TC1100010400.hg.1 | ENST00000379214.8 | Ensembl |
| Affymetrix Human Clariom D Assay | TC1100010400.hg.1 | ENST00000379214.8 | Ensembl |
| Affymetrix Human Clariom D Assay | TC1100010400.hg.1 | ENST00000379214.8 | Ensembl |
| Affymetrix Human Clariom D Assay | TC1100010400.hg.1 | ENST00000379214.8 | Ensembl |
| Affymetrix Human Clariom D Assay | TC1100010400.hg.1 | ENST00000379214.8 | Ensembl |
| Affymetrix Human Clariom D Assay | TC1100010400.hg.1 | ENST00000379214.8 | Ensembl |
| Affymetrix Human Clariom D Assay | TC1100010400.hg.1 | ENST00000379214.8 | Ensembl |
| Affymetrix Human Clariom D Assay | TC1100010400.hg.1 | ENST00000379214.8 | Ensembl |
| Affymetrix Human Clariom D Assay | TC1100010400.hg.1 | ENST00000379214.8 | Ensembl |
| Affymetrix Human Clariom D Assay | TC1100010400.hg.1 | ENST00000379214.8 | Ensembl |
| Affymetrix Human Clariom D Assay | TC1100010400.hg.1 | ENST00000379214.8 | Ensembl |
| Affymetrix Human Clariom D Assay | TC1100010400.hg.1 | ENST00000379214.8 | Ensembl |
| Affymetrix Human Clariom D Assay | TC1100010400.hg.1 | ENST00000379214.8 | Ensembl |
| Affymetrix Human Clariom D Assay | TC1100010400.hg.1 | ENST00000379214.8 | Ensembl |
| Affymetrix Human Clariom D Assay | TC1100010400.hg.1 | ENST00000379214.8 | Ensembl |
| Affymetrix Human Clariom D Assay | TC1100010400.hg.1 | ENST00000379214.8 | Ensembl |
| Affymetrix Human Clariom D Assay | TC1100010400.hg.1 | ENST00000379214.8 | Ensembl |
| Affymetrix Human Clariom D Assay | TC1100010400.hg.1 | ENST00000389858.4 | Ensembl |
| Affymetrix Human Clariom D Assay | TC1100010400.hg.1 | ENST00000389858.4 | Ensembl |
| Affymetrix Human Clariom D Assay | TC1100010400.hg.1 | ENST00000389858.4 | Ensembl |
| Affymetrix Human Clariom D Assay | TC1100010400.hg.1 | ENST00000389858.4 | Ensembl |
| Affymetrix Human Clariom D Assay | TC1100010400.hg.1 | ENST00000389858.4 | Ensembl |
| Affymetrix Human Clariom D Assay | TC1100010400.hg.1 | ENST00000389858.4 | Ensembl |
| Affymetrix Human Clariom D Assay | TC1100010400.hg.1 | ENST00000389858.4 | Ensembl |
| Affymetrix Human Clariom D Assay | TC1100010400.hg.1 | ENST00000389858.4 | Ensembl |
| Affymetrix Human Clariom D Assay | TC1100010400.hg.1 | ENST00000389858.4 | Ensembl |
| Affymetrix Human Clariom D Assay | TC1100010400.hg.1 | ENST00000389858.4 | Ensembl |
| Affymetrix Human Clariom D Assay | TC1100010400.hg.1 | ENST00000389858.4 | Ensembl |
| Affymetrix Human Clariom D Assay | TC1100010400.hg.1 | ENST00000389858.4 | Ensembl |
| Affymetrix Human Clariom D Assay | TC1100010400.hg.1 | ENST00000389858.4 | Ensembl |
| Affymetrix Human Clariom D Assay | TC1100010400.hg.1 | ENST00000389858.4 | Ensembl |
| Affymetrix Human Clariom D Assay | TC1100010400.hg.1 | ENST00000389858.4 | Ensembl |
| Affymetrix Human Clariom D Assay | TC1100010400.hg.1 | ENST00000389858.4 | Ensembl |
| Affymetrix Human Clariom D Assay | TC1100010400.hg.1 | ENST00000389858.4 | Ensembl |
| Affymetrix Human Clariom D Assay | TC1100010400.hg.1 | ENST00000389858.4 | Ensembl |
| Affymetrix Human Clariom D Assay | TC1100010400.hg.1 | ENST00000389858.4 | Ensembl |
| Affymetrix Human Clariom D Assay | TC1100010400.hg.1 | ENST00000389858.4 | Ensembl |
| Affymetrix Human Clariom D Assay | TC1100010400.hg.1 | ENST00000389858.4 | Ensembl |
| Affymetrix Human Clariom D Assay | TC1100010400.hg.1 | ENST00000389858.4 | Ensembl |
| Affymetrix Human Clariom D Assay | TC1100010400.hg.1 | ENST00000480977.2 | Ensembl |
| Affymetrix Human Clariom D Assay | TC1100010400.hg.1 | ENST00000480977.2 | Ensembl |
| Affymetrix Human Clariom D Assay | TC1100010400.hg.1 | ENST00000480977.2 | Ensembl |
| Affymetrix Human Clariom D Assay | TC1100010400.hg.1 | ENST00000489910.1 | Ensembl |
| Affymetrix Human Clariom D Assay | TC1100010400.hg.1 | ENST00000489910.1 | Ensembl |
| Affymetrix Human Clariom D Assay | TC1100010400.hg.1 | ENST00000489910.1 | Ensembl |
| Affymetrix Human Clariom D Assay | TC1100010400.hg.1 | ENST00000527773.1 | Ensembl |
| Affymetrix Human Clariom D Assay | TC1100010400.hg.1 | ENST00000527773.1 | Ensembl |
| Affymetrix Human Clariom D Assay | TC1100010400.hg.1 | ENST00000527773.1 | Ensembl |
| Affymetrix Human Clariom D Assay | TC1100010400.hg.1 | ENST00000527773.1 | Ensembl |
| Affymetrix Human Clariom D Assay | TC1100010400.hg.1 | ENST00000530658.1 | Ensembl |
| Affymetrix Human Clariom D Assay | TC1100010400.hg.1 | ENST00000530658.1 | Ensembl |
| Affymetrix Human Clariom D Assay | TC1100010400.hg.1 | ENST00000530658.1 | Ensembl |
| Affymetrix Human Clariom D Assay | TC1100010400.hg.1 | NM_018490.3 | Refseq |
| Affymetrix Human Clariom D Assay | TC1100010400.hg.1 | NM_018490.3 | Refseq |
| Affymetrix Human Clariom D Assay | TC1100010400.hg.1 | NM_018490.3 | Refseq |
| Affymetrix Human Clariom D Assay | TC1100010400.hg.1 | NM_018490.3 | Refseq |
| Affymetrix Human Clariom D Assay | TC1100010400.hg.1 | NM_018490.3 | Refseq |
| Affymetrix Human Clariom D Assay | TC1100010400.hg.1 | NM_018490.3 | Refseq |
| Affymetrix Human Clariom D Assay | TC1100010400.hg.1 | NM_018490.3 | Refseq |
| Affymetrix Human Clariom D Assay | TC1100010400.hg.1 | NM_018490.3 | Refseq |
| Affymetrix Human Clariom D Assay | TC1100010400.hg.1 | NM_018490.3 | Refseq |
| Affymetrix Human Clariom D Assay | TC1100010400.hg.1 | NM_018490.3 | Refseq |
| Affymetrix Human Clariom D Assay | TC1100010400.hg.1 | NM_018490.3 | Refseq |
| Affymetrix Human Clariom D Assay | TC1100010400.hg.1 | NM_018490.3 | Refseq |
| Affymetrix Human Clariom D Assay | TC1100010400.hg.1 | NM_018490.3 | Refseq |
| Affymetrix Human Clariom D Assay | TC1100010400.hg.1 | NM_018490.3 | Refseq |
| Affymetrix Human Clariom D Assay | TC1100010400.hg.1 | NM_018490.3 | Refseq |
| Affymetrix Human Clariom D Assay | TC1100010400.hg.1 | NM_018490.3 | Refseq |
| Affymetrix Human Clariom D Assay | TC1100010400.hg.1 | NM_018490.3 | Refseq |
| Affymetrix Human Clariom D Assay | TC1100010400.hg.1 | NM_018490.3 | Refseq |
| Affymetrix Human Clariom D Assay | TC1100010400.hg.1 | NM_018490.3 | Refseq |
| Affymetrix Human Clariom D Assay | TC1100010400.hg.1 | NM_018490.3 | Refseq |
| Affymetrix Human Clariom D Assay | TC1100010400.hg.1 | NM_018490.3 | Refseq |
| Affymetrix Human Clariom D Assay | TC1100010400.hg.1 | NM_018490.3 | Refseq |
| Affymetrix Human Clariom D Assay | TC1100010400.hg.1 | NM_018490.3 | Refseq |
| Affymetrix Human Clariom D Assay | PSR1100178836.hg.1 | ENST00000379214.8 | Ensembl |
| Affymetrix Human Clariom D Assay | PSR1100178836.hg.1 | ENST00000389858.4 | Ensembl |
| Affymetrix Human Clariom D Assay | PSR1100178836.hg.1 | NM_018490.3 | Refseq |
| Affymetrix Human Clariom D Assay | PSR1100178836.hg.1 | ENST00000379214.8 | Ensembl |
| Affymetrix Human Clariom D Assay | PSR1100178836.hg.1 | ENST00000389858.4 | Ensembl |
| Affymetrix Human Clariom D Assay | PSR1100178836.hg.1 | NM_018490.3 | Refseq |
| Affymetrix Human Clariom D Assay | PSR1100178836.hg.1 | TC1100010400.hg.1 | Clariom D TC |

LGR4
Genomic Map