Assay Details
| Gene Symbol: ZCCHC6 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | NM_001185059.1 | NP_001171988.1 | 15 - 16 | 3252 | 122 | |
| NM_024617.3 | NP_078893.2 | 15 - 16 | 3362 | 122 | ||
| XM_005252207.3 | XP_005252264.1 | 15 - 16 | 3314 | 122 | ||
| XM_005252208.2 | XP_005252265.1 | 15 - 16 | 3314 | 122 | ||
| XM_006717283.1 | XP_006717346.1 | 15 - 16 | 3314 | 122 | ||
| XM_011519011.2 | XP_011517313.1 | 15 - 16 | 3314 | 122 | ||
| XM_011519012.2 | XP_011517314.1 | 15 - 16 | 3262 | 122 | ||
| XM_011519013.2 | XP_011517315.1 | 15 - 16 | 3931 | 122 | ||
| XM_011519014.2 | XP_011517316.1 | 15 - 16 | 3533 | 122 | ||
| XM_011519015.1 | XP_011517317.1 | 15 - 16 | 3314 | 122 | ||
| XM_011519016.1 | XP_011517318.1 | 15 - 16 | 3314 | 122 | ||
| XM_017015130.1 | XP_016870619.1 | 15 - 16 | 3930 | 122 | ||
| XM_017015131.1 | XP_016870620.1 | 15 - 16 | 3572 | 122 | ||
| XM_017015132.1 | XP_016870621.1 | 16 - 17 | 3477 | 122 | ||
| GenBank mRNA | AB051498.1 | - | 7 - 8 | 1900 | 122 | |
| AK023471.1 | - | 1 - 2 | 62 | 122 | ||
| AK055948.1 | - | 3 - 4 | 1212 | 122 | ||
| AL832193.1 | - | 16 - 17 | 3404 | 122 | ||
| AL832988.1 | - | 2 - 3 | 200 | 122 | ||
| BC144523.1 | - | 14 - 15 | 3150 | 122 | ||
| BX641077.1 | - | 14 - 15 | 4712 | 122 | ||
| CR933643.1 | - | 15 - 16 | 3226 | 122 | ||
| CR933644.1 | - | 15 - 16 | 3804 | 122 | ||
More Information
Updated Mapping Information
This assay targets the same location as HTA transcript cluster(s) TC09001293.hg.1
Additional Information:
This assay is in the 5'UTR region of the AK023471.1 transcripts.
Set Membership: |
|

ZCCHC6
Genomic Map