| Array Name |
Probe IDs |
Targets |
Target Type |
| Affymetrix Human Clariom D Assay |
PSR0300184485.hg.1 |
ENST00000351747.6 |
Ensembl |
| Affymetrix Human Clariom D Assay |
PSR0300184485.hg.1 |
ENST00000495027.5 |
Ensembl |
| Affymetrix Human Clariom D Assay |
PSR0300184485.hg.1 |
NM_001291661.1 |
Refseq |
| Affymetrix Human Clariom D Assay |
PSR0300184485.hg.1 |
ENST00000351747.6 |
Ensembl |
| Affymetrix Human Clariom D Assay |
PSR0300184485.hg.1 |
ENST00000495027.5 |
Ensembl |
| Affymetrix Human Clariom D Assay |
PSR0300184485.hg.1 |
NM_001291661.1 |
Refseq |
| Affymetrix Human Clariom D Assay |
PSR0300184485.hg.1 |
TC0300011267.hg.1 |
Clariom D TC |