Assay Details
| Gene Symbol: PDE4D | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | NM_001104631.1 | NP_001098101.1 | 10 - 11 | 1619 | 77 | |
| NM_001165899.1 | NP_001159371.1 | 12 - 13 | 1510 | 77 | ||
| NM_001197218.1 | NP_001184147.1 | 10 - 11 | 1358 | 77 | ||
| NM_001197219.1 | NP_001184148.1 | 10 - 11 | 1136 | 77 | ||
| NM_001197220.1 | NP_001184149.1 | 10 - 11 | 1774 | 77 | ||
| NM_001197221.1 | NP_001184150.1 | 6 - 7 | 1094 | 77 | ||
| NM_001197222.1 | NP_001184151.1 | 5 - 6 | 1180 | 77 | ||
| NM_001197223.1 | NP_001184152.1 | 5 - 6 | 970 | 77 | ||
| NM_006203.4 | NP_006194.2 | 10 - 11 | 1162 | 77 | ||
| XM_005248537.3 | XP_005248594.1 | 10 - 11 | 2316 | 77 | ||
| XM_005248538.4 | XP_005248595.1 | 11 - 12 | 4217 | 77 | ||
| XM_011543469.1 | XP_011541771.1 | 13 - 14 | 1599 | 77 | ||
| XM_011543470.2 | XP_011541772.1 | 13 - 14 | 1523 | 77 | ||
| XM_011543471.1 | XP_011541773.1 | 12 - 13 | 1476 | 77 | ||
| XM_011543473.1 | XP_011541775.1 | 12 - 13 | 1424 | 77 | ||
| XM_011543477.2 | XP_011541779.1 | 12 - 13 | 1426 | 77 | ||
| XM_017009565.1 | XP_016865054.1 | 13 - 14 | 1636 | 77 | ||
| XM_017009566.1 | XP_016865055.1 | 13 - 14 | 1526 | 77 | ||
| XM_017009567.1 | XP_016865056.1 | 11 - 12 | 1264 | 77 | ||
| XM_017009568.1 | XP_016865057.1 | 13 - 14 | 1510 | 77 | ||
| XM_017009569.1 | XP_016865058.1 | 12 - 13 | 1389 | 77 | ||
| XM_017009570.1 | XP_016865059.1 | 9 - 10 | 2209 | 77 | ||
| XM_017009571.1 | XP_016865060.1 | 12 - 13 | 1432 | 77 | ||
| GenBank mRNA | AF012073.1 | - | 10 - 11 | 1372 | 77 | |
| AF012074.1 | - | 6 - 7 | 1108 | 77 | ||
| AF536975.1 | - | 5 - 6 | 777 | 77 | ||
| AF536976.1 | - | 12 - 13 | 1333 | 77 | ||
| AF536977.1 | - | 10 - 11 | 1136 | 77 | ||
| AJ250852.1 | - | 6 - 7 | 704 | 77 | ||
| AJ250854.1 | - | 11 - 12 | 1299 | 77 | ||
| AY245866.1 | - | 12 - 13 | 1475 | 77 | ||
| AY245867.1 | - | 10 - 11 | 1774 | 77 | ||
| AY388960.1 | - | 10 - 11 | 1057 | 77 | ||
| BC036319.1 | - | 5 - 6 | 972 | 4828512 | 77 | |
| L20970.1 | - | 10 - 11 | 1170 | 77 | ||
| U02882.1 | - | 9 - 10 | 940 | 77 | ||
| U50157.1 | - | 5 - 6 | 778 | 77 | ||
| U50158.1 | - | 6 - 7 | 692 | 77 | ||
| U50159.1 | - | 10 - 11 | 1113 | 77 | ||
More Information
Set Membership: |
|

PDE4D
Genomic Map