Assay Details
| Gene Symbol: GUCY2GP | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | NR_028134.1 | 2 - 3 | 710 | 145 | ||
More Information
Updated Mapping Information
This assay targets the same location as HTA transcript cluster(s) TC10001664.hg.1
Set Membership: |
|
MicroArray Probes:
Affymetrix
| Array Name | Probe IDs | Targets | Target Type |
|---|---|---|---|
| Affymetrix Human Transcriptome Array 2.0 | PSR10021849.hg.1 | NR_028134.1 | Refseq |
| Affymetrix Human Transcriptome Array 2.0 | PSR10021849.hg.1 | TC10001664.hg.1 | HTA TC |
| Affymetrix Human Transcriptome Array 2.0 | TC1000011842.hg.1 | ENST00000479705.5 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC1000011842.hg.1 | ENST00000479705.5 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC1000011842.hg.1 | ENST00000479705.5 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC1000011842.hg.1 | ENST00000479705.5 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC1000011842.hg.1 | ENST00000479705.5 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC1000011842.hg.1 | ENST00000479705.5 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC1000011842.hg.1 | ENST00000479705.5 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC1000011842.hg.1 | ENST00000479705.5 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC1000011842.hg.1 | ENST00000479705.5 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC1000011842.hg.1 | ENST00000479705.5 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC1000011842.hg.1 | ENST00000479705.5 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC1000011842.hg.1 | ENST00000479705.5 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC1000011842.hg.1 | ENST00000479705.5 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC1000011842.hg.1 | ENST00000479705.5 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC1000011842.hg.1 | ENST00000479705.5 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC1000011842.hg.1 | ENST00000479705.5 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC1000011842.hg.1 | ENST00000479705.5 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC1000011842.hg.1 | ENST00000479705.5 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC1000011842.hg.1 | ENST00000479705.5 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC1000011842.hg.1 | ENST00000479705.5 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC1000011842.hg.1 | ENST00000479705.5 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC1000011842.hg.1 | ENST00000479705.5 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC1000011842.hg.1 | ENST00000479705.5 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC1000011842.hg.1 | ENST00000479705.5 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC1000011842.hg.1 | ENST00000479705.5 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC1000011842.hg.1 | ENST00000479705.5 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC1000011842.hg.1 | ENST00000490269.1 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC1000011842.hg.1 | ENST00000490269.1 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC1000011842.hg.1 | ENST00000490269.1 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC1000011842.hg.1 | ENST00000490269.1 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC1000011842.hg.1 | ENST00000490269.1 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC1000011842.hg.1 | ENST00000490269.1 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC1000011842.hg.1 | ENST00000490269.1 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC1000011842.hg.1 | ENST00000490269.1 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC1000011842.hg.1 | ENST00000490269.1 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC1000011842.hg.1 | ENST00000490269.1 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC1000011842.hg.1 | ENST00000490269.1 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC1000011842.hg.1 | ENST00000490269.1 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC1000011842.hg.1 | ENST00000490269.1 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC1000011842.hg.1 | ENST00000490269.1 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC1000011842.hg.1 | NR_028134.1 | Refseq |
| Affymetrix Human Transcriptome Array 2.0 | TC1000011842.hg.1 | NR_028134.1 | Refseq |
| Affymetrix Human Transcriptome Array 2.0 | TC1000011842.hg.1 | NR_028134.1 | Refseq |
| Affymetrix Human Transcriptome Array 2.0 | TC1000011842.hg.1 | NR_028134.1 | Refseq |
| Affymetrix Human Transcriptome Array 2.0 | TC1000011842.hg.1 | NR_028134.1 | Refseq |
| Affymetrix Human Transcriptome Array 2.0 | TC1000011842.hg.1 | NR_028134.1 | Refseq |
| Affymetrix Human Transcriptome Array 2.0 | TC1000011842.hg.1 | NR_028134.1 | Refseq |
| Affymetrix Human Transcriptome Array 2.0 | TC1000011842.hg.1 | NR_028134.1 | Refseq |
| Affymetrix Human Transcriptome Array 2.0 | TC1000011842.hg.1 | NR_028134.1 | Refseq |
| Affymetrix Human Transcriptome Array 2.0 | TC1000011842.hg.1 | NR_028134.1 | Refseq |
| Affymetrix Human Transcriptome Array 2.0 | TC1000011842.hg.1 | NR_028134.1 | Refseq |
| Affymetrix Human Transcriptome Array 2.0 | TC1000011842.hg.1 | NR_028134.1 | Refseq |
| Affymetrix Human Transcriptome Array 2.0 | TC1000011842.hg.1 | NR_028134.1 | Refseq |
| Affymetrix Human Transcriptome Array 2.0 | TC1000011842.hg.1 | NR_028134.1 | Refseq |
| Affymetrix Human Transcriptome Array 2.0 | TC1000011842.hg.1 | NR_028134.1 | Refseq |
| Affymetrix Human Transcriptome Array 2.0 | TC1000011842.hg.1 | NR_028134.1 | Refseq |
| Affymetrix Human Transcriptome Array 2.0 | TC1000011842.hg.1 | NR_028134.1 | Refseq |
| Affymetrix Human Transcriptome Array 2.0 | TC1000011842.hg.1 | NR_028134.1 | Refseq |
| Affymetrix Human Transcriptome Array 2.0 | TC1000011842.hg.1 | NR_028134.1 | Refseq |
| Affymetrix Human Transcriptome Array 2.0 | TC1000011842.hg.1 | NR_028134.1 | Refseq |
| Affymetrix Human Transcriptome Array 2.0 | TC1000011842.hg.1 | NR_028134.1 | Refseq |
| Affymetrix Human Transcriptome Array 2.0 | TC1000011842.hg.1 | NR_028134.1 | Refseq |
| Affymetrix Human Transcriptome Array 2.0 | TC1000011842.hg.1 | NR_028134.1 | Refseq |
| Affymetrix Human Transcriptome Array 2.0 | TC1000011842.hg.1 | NR_028134.1 | Refseq |
| Affymetrix Human Transcriptome Array 2.0 | PSR10021847.hg.1 | NR_028134.1 | Refseq |
| Affymetrix Human Transcriptome Array 2.0 | PSR10021847.hg.1 | TC10001664.hg.1 | HTA TC |
| Affymetrix Human Transcriptome Array 2.0 | PSR10021848.hg.1 | NR_028134.1 | Refseq |
| Affymetrix Human Transcriptome Array 2.0 | PSR10021848.hg.1 | TC10001664.hg.1 | HTA TC |
| Affymetrix Human Clariom D Assay | TC1000011842.hg.1 | ENST00000479705.5 | Ensembl |
| Affymetrix Human Clariom D Assay | TC1000011842.hg.1 | ENST00000479705.5 | Ensembl |
| Affymetrix Human Clariom D Assay | TC1000011842.hg.1 | ENST00000479705.5 | Ensembl |
| Affymetrix Human Clariom D Assay | TC1000011842.hg.1 | ENST00000479705.5 | Ensembl |
| Affymetrix Human Clariom D Assay | TC1000011842.hg.1 | ENST00000479705.5 | Ensembl |
| Affymetrix Human Clariom D Assay | TC1000011842.hg.1 | ENST00000479705.5 | Ensembl |
| Affymetrix Human Clariom D Assay | TC1000011842.hg.1 | ENST00000479705.5 | Ensembl |
| Affymetrix Human Clariom D Assay | TC1000011842.hg.1 | ENST00000479705.5 | Ensembl |
| Affymetrix Human Clariom D Assay | TC1000011842.hg.1 | ENST00000479705.5 | Ensembl |
| Affymetrix Human Clariom D Assay | TC1000011842.hg.1 | ENST00000479705.5 | Ensembl |
| Affymetrix Human Clariom D Assay | TC1000011842.hg.1 | ENST00000479705.5 | Ensembl |
| Affymetrix Human Clariom D Assay | TC1000011842.hg.1 | ENST00000479705.5 | Ensembl |
| Affymetrix Human Clariom D Assay | TC1000011842.hg.1 | ENST00000479705.5 | Ensembl |
| Affymetrix Human Clariom D Assay | TC1000011842.hg.1 | ENST00000479705.5 | Ensembl |
| Affymetrix Human Clariom D Assay | TC1000011842.hg.1 | ENST00000479705.5 | Ensembl |
| Affymetrix Human Clariom D Assay | TC1000011842.hg.1 | ENST00000479705.5 | Ensembl |
| Affymetrix Human Clariom D Assay | TC1000011842.hg.1 | ENST00000479705.5 | Ensembl |
| Affymetrix Human Clariom D Assay | TC1000011842.hg.1 | ENST00000479705.5 | Ensembl |
| Affymetrix Human Clariom D Assay | TC1000011842.hg.1 | ENST00000479705.5 | Ensembl |
| Affymetrix Human Clariom D Assay | TC1000011842.hg.1 | ENST00000479705.5 | Ensembl |
| Affymetrix Human Clariom D Assay | TC1000011842.hg.1 | ENST00000479705.5 | Ensembl |
| Affymetrix Human Clariom D Assay | TC1000011842.hg.1 | ENST00000479705.5 | Ensembl |
| Affymetrix Human Clariom D Assay | TC1000011842.hg.1 | ENST00000479705.5 | Ensembl |
| Affymetrix Human Clariom D Assay | TC1000011842.hg.1 | ENST00000479705.5 | Ensembl |
| Affymetrix Human Clariom D Assay | TC1000011842.hg.1 | ENST00000479705.5 | Ensembl |
| Affymetrix Human Clariom D Assay | TC1000011842.hg.1 | ENST00000479705.5 | Ensembl |
| Affymetrix Human Clariom D Assay | TC1000011842.hg.1 | ENST00000490269.1 | Ensembl |
| Affymetrix Human Clariom D Assay | TC1000011842.hg.1 | ENST00000490269.1 | Ensembl |
| Affymetrix Human Clariom D Assay | TC1000011842.hg.1 | ENST00000490269.1 | Ensembl |
| Affymetrix Human Clariom D Assay | TC1000011842.hg.1 | ENST00000490269.1 | Ensembl |
| Affymetrix Human Clariom D Assay | TC1000011842.hg.1 | ENST00000490269.1 | Ensembl |
| Affymetrix Human Clariom D Assay | TC1000011842.hg.1 | ENST00000490269.1 | Ensembl |
| Affymetrix Human Clariom D Assay | TC1000011842.hg.1 | ENST00000490269.1 | Ensembl |
| Affymetrix Human Clariom D Assay | TC1000011842.hg.1 | ENST00000490269.1 | Ensembl |
| Affymetrix Human Clariom D Assay | TC1000011842.hg.1 | ENST00000490269.1 | Ensembl |
| Affymetrix Human Clariom D Assay | TC1000011842.hg.1 | ENST00000490269.1 | Ensembl |
| Affymetrix Human Clariom D Assay | TC1000011842.hg.1 | ENST00000490269.1 | Ensembl |
| Affymetrix Human Clariom D Assay | TC1000011842.hg.1 | ENST00000490269.1 | Ensembl |
| Affymetrix Human Clariom D Assay | TC1000011842.hg.1 | ENST00000490269.1 | Ensembl |
| Affymetrix Human Clariom D Assay | TC1000011842.hg.1 | ENST00000490269.1 | Ensembl |
| Affymetrix Human Clariom D Assay | TC1000011842.hg.1 | NR_028134.1 | Refseq |
| Affymetrix Human Clariom D Assay | TC1000011842.hg.1 | NR_028134.1 | Refseq |
| Affymetrix Human Clariom D Assay | TC1000011842.hg.1 | NR_028134.1 | Refseq |
| Affymetrix Human Clariom D Assay | TC1000011842.hg.1 | NR_028134.1 | Refseq |
| Affymetrix Human Clariom D Assay | TC1000011842.hg.1 | NR_028134.1 | Refseq |
| Affymetrix Human Clariom D Assay | TC1000011842.hg.1 | NR_028134.1 | Refseq |
| Affymetrix Human Clariom D Assay | TC1000011842.hg.1 | NR_028134.1 | Refseq |
| Affymetrix Human Clariom D Assay | TC1000011842.hg.1 | NR_028134.1 | Refseq |
| Affymetrix Human Clariom D Assay | TC1000011842.hg.1 | NR_028134.1 | Refseq |
| Affymetrix Human Clariom D Assay | TC1000011842.hg.1 | NR_028134.1 | Refseq |
| Affymetrix Human Clariom D Assay | TC1000011842.hg.1 | NR_028134.1 | Refseq |
| Affymetrix Human Clariom D Assay | TC1000011842.hg.1 | NR_028134.1 | Refseq |
| Affymetrix Human Clariom D Assay | TC1000011842.hg.1 | NR_028134.1 | Refseq |
| Affymetrix Human Clariom D Assay | TC1000011842.hg.1 | NR_028134.1 | Refseq |
| Affymetrix Human Clariom D Assay | TC1000011842.hg.1 | NR_028134.1 | Refseq |
| Affymetrix Human Clariom D Assay | TC1000011842.hg.1 | NR_028134.1 | Refseq |
| Affymetrix Human Clariom D Assay | TC1000011842.hg.1 | NR_028134.1 | Refseq |
| Affymetrix Human Clariom D Assay | TC1000011842.hg.1 | NR_028134.1 | Refseq |
| Affymetrix Human Clariom D Assay | TC1000011842.hg.1 | NR_028134.1 | Refseq |
| Affymetrix Human Clariom D Assay | TC1000011842.hg.1 | NR_028134.1 | Refseq |
| Affymetrix Human Clariom D Assay | TC1000011842.hg.1 | NR_028134.1 | Refseq |
| Affymetrix Human Clariom D Assay | TC1000011842.hg.1 | NR_028134.1 | Refseq |
| Affymetrix Human Clariom D Assay | TC1000011842.hg.1 | NR_028134.1 | Refseq |
| Affymetrix Human Clariom D Assay | TC1000011842.hg.1 | NR_028134.1 | Refseq |
| Affymetrix Human Clariom D Assay | PSR1000181119.hg.1 | ENST00000479705.5 | Ensembl |
| Affymetrix Human Clariom D Assay | PSR1000181119.hg.1 | NR_028134.1 | Refseq |
| Affymetrix Human Clariom D Assay | PSR1000181119.hg.1 | ENST00000479705.5 | Ensembl |
| Affymetrix Human Clariom D Assay | PSR1000181119.hg.1 | NR_028134.1 | Refseq |
| Affymetrix Human Clariom D Assay | PSR1000181119.hg.1 | TC1000011842.hg.1 | Clariom D TC |
| Affymetrix Human Clariom D Assay | PSR1000181120.hg.1 | ENST00000479705.5 | Ensembl |
| Affymetrix Human Clariom D Assay | PSR1000181120.hg.1 | NR_028134.1 | Refseq |
| Affymetrix Human Clariom D Assay | PSR1000181120.hg.1 | ENST00000479705.5 | Ensembl |
| Affymetrix Human Clariom D Assay | PSR1000181120.hg.1 | NR_028134.1 | Refseq |
| Affymetrix Human Clariom D Assay | PSR1000181120.hg.1 | TC1000011842.hg.1 | Clariom D TC |
| Affymetrix Human Clariom D Assay | PSR1000181118.hg.1 | ENST00000479705.5 | Ensembl |
| Affymetrix Human Clariom D Assay | PSR1000181118.hg.1 | NR_028134.1 | Refseq |
| Affymetrix Human Clariom D Assay | PSR1000181118.hg.1 | ENST00000479705.5 | Ensembl |
| Affymetrix Human Clariom D Assay | PSR1000181118.hg.1 | NR_028134.1 | Refseq |
| Affymetrix Human Clariom D Assay | PSR1000181118.hg.1 | TC1000011842.hg.1 | Clariom D TC |

GUCY2GP
Genomic Map